Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 400
Cole-Parmer Disposable in-Line Ga
to
Cole-Parmer Dissolved Oxygen Process Instruments
Image
Product Detail
More Detail
Cole-Parmer Disposable in-Line Gas/Liquid Filters
Buy Cole-Parmer Disposable in-Line Gas/Liquid Filters Items Including:
Disposable In-Line Gas/Liquid Filter; Nylon with 1/2" OD connection; g (YO-02909-17)
Disposable In-Line Gas/Liquid Filter; Nylon with 1/2" OD connection; g (YO-02909-37).
More Details
Cole-Parmer Disposable in-Line Mixers
Buy Cole-Parmer Disposable in-Line Mixers Items Including:
Mounted in-line mixer; 5/16" tube OD, 12 elements, 10/pack (YO-04668-02)
Mounted in-line mixer; 3/8" tube OD, 12 elements, 10/pack (YO-04668-04)
Mounted in-line mixer; 1/2" tube OD, 12 elements, 10/pack (YO-04668-06).
More Details
Cole-Parmer Disposable Lab Containers
Buy Cole-Parmer Disposable Lab Containers Items Including:
Cole-Parmer Disposable Lab Container, Polypropylene Copolymer (PPCO), (YO-06727-00)
Cole-Parmer Disposable Lab Container, Polypropylene Copolymer (PPCO), (YO-06727-10).
More Details
Cole-Parmer Disposable Lab Containers, Ppco
Buy Cole-Parmer Disposable Lab Containers, Ppco Items Including:
Cole-Parmer Disposable Lab Container, Polypropylene Copolymer (PPCO), (YO-06727-00)
Cole-Parmer Disposable Lab Container, Polypropylene Copolymer (PPCO), (YO-06727-10).
More Details
Cole-Parmer Disposable Micropipettes
Buy Cole-Parmer Disposable Micropipettes Items Including:
Disposable Micropipettes, 0.25 µL, 100/pk (1-000-00025)
Disposable Micropipettes, 0.5 µL, 100/pk (1-000-0005)
Disposable Micropipettes, 10 µL, 100/pk (1-000-0100)
Disposable Micropipettes, 100 µL, 100/pk (1-000-1000).
More Details
Cole-Parmer Disposable Milling Jars
Buy Cole-Parmer Disposable Milling Jars Items Including:
HDPE milling jar; 0.125 gal
(0.5 L)
HDPE milling jar, 0.25 gal
(0.9 L)
HDPE milling jar, 0.5 gal
More Details
Cole-Parmer Disposable Pipette Tips
Buy Cole-Parmer Disposable Pipette Tips Items Including:
Cole-Parmer Disposable Pipette Tips, sterile, 2" L, 0-200 µL, 960/c (A23091PRX)
Cole-Parmer Disposable Pipette Tips, sterile, 2" L, 0-200 µL, 960/c (A23071PRX)
Cole-Parmer Disposable Pipette Tips, non-sterile, 3-1/8" L, 200-1300.
More Details
Cole-Parmer Disposable Plastic Tip
Buy Cole-Parmer Disposable Plastic Tip Items Including:
LabGEN Tip Probes, 0.25 to 50 mL sample volume, Hard Tissue, 25/pack
LabGEN Tip Probes, 0.25 to 50 mL, Hard Tissue, 50/pack (YO-04727-85)
Eight-channel plastic aspirator for disposable tips with ejection devi.
More Details
Cole-Parmer Disposable Plastic Tip Homogenizer Probes
Buy Cole-Parmer Disposable Plastic Tip Homogenizer Probes Items Including:
LabGEN Tip Probes, 0.25 to 50 mL sample volume, Hard Tissue, 25/pack
LabGEN Tip Probes, 0.25 to 50 mL, Hard Tissue, 50/pack (YO-04727-85)
LabGEN Tip Probes, 0.25 to 50 mL, Soft Tissue, 25/pack (YO-04727-86).
More Details
Cole-Parmer Disposable Serological Pipettes
Buy Cole-Parmer Disposable Serological Pipettes Items Including:
Disposable Pasteur Pipettes, Plugged End Borosilicate Glass, 145 mm, 1 (63A53P)
Disposable Pasteur Pipettes, Plugged End Borosilicate Glass, 230 mm, 1 (63B92P)
Disposable Pasteur Pipettes, Plugged End Soda Lime Glass, 145 mm, 1000 (63A54P).
More Details
Cole-Parmer Disposable Stir Bars
Buy Cole-Parmer Disposable Stir Bars Items Including:
1/2" X 1/8" Disposable Stir Bars (80037-1065)
1" X 5/16" Disposable Stir Bars (80037-1066)
1-1/2" X 5/16" Disposable Stir Bars (80037-1067).
More Details
Cole-Parmer Disposable Test/Culture Tubes, Pp
Buy Cole-Parmer Disposable Test/Culture Tubes, Pp Items Including:
Disposable polypropylene test/culture tubes, 20 mL
Disposable polypropylene test/culture tubes, 25 mL.
More Details
Cole-Parmer Disposable Tips
Buy Cole-Parmer Disposable Tips Items Including:
Cole-Parmer Disposable Pipette Tips, sterile, 2" L, 0-200 µL, 960/c (A23091PRX)
Cole-Parmer Disposable Pipette Tips, sterile, 2" L, 0-200 µL, 960/c (A23071PRX)
Cole-Parmer Disposable Pipette Tips, non-sterile, 3-1/8" L, 200-1300.
More Details
Cole-Parmer Disposable Tips for Macropipettors
Buy Cole-Parmer Disposable Tips for Macropipettors Items Including:
Cole-Parmer Green Pipette Tip, 1000-5000 µL, 200/pk
Disposable Tips, 0.4-2 mL, 200/bag.
More Details
Cole-Parmer Disposable Transfer Pipettes
Buy Cole-Parmer Disposable Transfer Pipettes Items Including:
Transfer Pipette, 1.5ml, Graduated, 500/pk (YO-06226-37)
Transfer Pipette, 3.9ml, Graduated, 500/pk (YO-06226-41)
Transfer Pipette, 5.8ml, Graduated, Large Bulb, 500/pk (YO-06226-61)
Transfer Pipette 7.5ml Graduated Large Bulb 500/pk (YO-06226-63).
More Details
Cole-Parmer Disposable Uv Cuvettes
Buy Cole-Parmer Disposable Uv Cuvettes Items Including:
Disposable UV cuvettes, methacrylate; 4.5 mL capacity, 500/pack (302235-0002)
Disposable UV cuvettes, methacrylate; 1.5 mL capacity, 500/pack (302225-0002).
More Details
Cole-Parmer Disposable Visible Cuvettes
Buy Cole-Parmer Disposable Visible Cuvettes Items Including:
Disposable Visible Cuvettes, polystyrene; 4.5 mL capacity, 500/pack (202295)
Disposable UV cuvettes, methacrylate; 4.5 mL capacity, 500/pack (302235-0002)
Disposable UV cuvettes, methacrylate; 1.5 mL capacity, 500/pack (302225-0002).
More Details
Cole-Parmer Disposal Bags
Buy Cole-Parmer Disposal Bags Items Including:
High-Molecular Weight Bags, 16 gallon
High-Molecular Weight Bags, 30 gallon
High-Molecular Weight Bags, 33 gallon
High-Molecular Weight Bags, 56 gallon
50 mL self-standing tube, PPCO, Sterile, 20 bags of 25 (3262)
More Details
Cole-Parmer Disposal Boxes
Buy Cole-Parmer Disposal Boxes Items Including:
Glass Disposal Boxes, Bench Style, 6 per pack
Glass Disposal Boxes, Intermediate Style, 6 per pack
Glass Disposal Boxes, Floor Style, 6 per pack
Plastic Disposal Box, Floor Style, 6 per pack
Disposable Hemacytometers 50/Box - Fuchs Rosenthal Grid.
More Details
Cole-Parmer Disposal Cans
Buy Cole-Parmer Disposal Cans Items Including:
Cole-Parmer Disposable Lab Container, Polypropylene Copolymer (PPCO), (YO-06727-00)
Cole-Parmer Disposable Lab Container, Polypropylene Copolymer (PPCO), (YO-06727-10)
Cole-Parmer Disposable Pipette Tips, 1-13/16" L, 0.2-10 µL, 10/00/c.
More Details
Cole-Parmer Dissecting Instruments and Kits
Buy Cole-Parmer Dissecting Instruments and Kits Items Including:
Dissecting T-Pins; 1-1/2 inch, 1000/CS (YO-10915-03).
More Details
Cole-Parmer Dissecting Kit
Buy Cole-Parmer Dissecting Kit Items Including:
Dissecting T-Pins; 1-1/2 inch, 1000/CS (YO-10915-03).
More Details
Cole-Parmer Dissecting Trays
Buy Cole-Parmer Dissecting Trays Items Including:
Cole-Parmer Dissecting Tray, 11.5" x 7.5" x 2" (YO-10915-12).
More Details
Cole-Parmer Dissection and Surgical Instruments
Buy Cole-Parmer Dissection and Surgical Instruments Items Including:
Dissecting T-Pins; 1-1/2 inch, 1000/CS (YO-10915-03)
Plain Gut Sterile Sutures, 13 mm, Needle: C-3 (YO-10901-00)
Plain Gut Sterile Sutures, 13 mm, Needle: P-3 (YO-10901-02)
Plain Gut Sterile Sutures, 19 mm, Needle: FS-2 3-0 27" (YO-10901-04).
More Details
Cole-Parmer Dissection T-Pins
Buy Cole-Parmer Dissection T-Pins Items Including:
Dissecting T-Pins; 1-1/2 inch, 1000/CS (YO-10915-03).
More Details
Cole-Parmer Dissolved Oxygen Benchtop Meters
Buy Cole-Parmer Dissolved Oxygen Benchtop Meters Items Including:
Cole-Parmer Lab Dissolved Oxygen Meter, Digital
Cole-Parmer Field Dissolved Oxygen Meter, Digital
Strathkelvin Precision Dissolved Oxygen Respirometer, two-channel
Strathkelvin Precision Dissolved Oxygen Respirometer, six-channel.
More Details
Cole-Parmer Dissolved Oxygen Control
Buy Cole-Parmer Dissolved Oxygen Control Items Including:
Benchtop Dissolved Oxygen Meter, 110 VAC
Fill solution for chlorine dioxide sensors, 30-mL bottle (YO-99673-11)
Fill solution for Free Chlorine sensors, 30-mL bottle (YO-99673-13)
Replacement membrane cap for Chlorine Dioxide sensors (YO-99673-15).
More Details
Cole-Parmer Dissolved Oxygen Meters
Buy Cole-Parmer Dissolved Oxygen Meters Items Including:
Cole-Parmer Field Dissolved Oxygen Meter, Digital
NIST-Traceable calibration for dissolved oxygen meters
Cole-Parmer Glass Body Fermentation DO Probe, 180mm immersion depth, p
Cole-Parmer Glass Body Fermentation DO Probe, 210mm immersion depth, p.
More Details
Cole-Parmer Dissolved Oxygen Probes and Sensors
Buy Cole-Parmer Dissolved Oxygen Probes and Sensors Items Including:
Cole-Parmer Glass Body Fermentation DO Probe, 180mm immersion depth, p
Cole-Parmer Stainless Steel Body Fermentation DO Probe, 150-mm immersi (YO-05726-00)
Cole-Parmer Stainless Steel Body Fermentation DO Probe, 320-mm immersi (YO-05726-02).
More Details
Cole-Parmer Dissolved Oxygen Process Instruments
Buy Cole-Parmer Dissolved Oxygen Process Instruments Items Including:
Benchtop Dissolved Oxygen Meter, 110 VAC
Fill solution for chlorine dioxide sensors, 30-mL bottle (YO-99673-11)
Fill solution for Free Chlorine sensors, 30-mL bottle (YO-99673-13)
Replacement membrane cap for Chlorine Dioxide sensors (YO-99673-15).
More Details
70618
results
<<<Previous
396
397
398
399
400
401
402
403
404
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
310
320
330
340
350
360
370
380
385
386
387
388
389
390
391
392
393
394
395
396
397
398
399
400
401
402
403
404
405
406
407
408
409
410
411
412
413
414
420
430
440
450
460
470
480
490
500
600
700
800
900
1,000
1,100
1,200
1,300
2,000
2,354
Cole-Parmer Disposable in-Line Ga to Cole-Parmer Dissolved Oxygen Process Instruments
>
31125329-s-toc_2 31125329stoc2