Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 60
Air Powered Lab Stirrer
to
Air Pump Inlet Outlet
Image
Product Detail
More Detail
Air Powered Lab Stirrer
Buy Air Powered Lab Stirrer Items Including:
Cole-Parmer Battery-Powered Magnetic Stirrer
Bulk container mixer; 1/2 hp, 350 rpm, air powered, requires separate
Low-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM (YO-04680-00)
Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00).
More Details
Air Powered Mixer from Mcmaster Carr
Buy Air Powered Mixer from Mcmaster Carr Items Including:
Low-cost, gear-drive air motor mixer; 8-170 rpm (YO-04681-00)
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00)
Lightnin general-purpose air-drive mixer.
More Details
Air Powered Mixer Plastic
Buy Air Powered Mixer Plastic Items Including:
Low-cost, gear-drive air motor mixer; 8-170 rpm (YO-04681-00)
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00)
Lightnin general-purpose air-drive mixer.
More Details
Air Powered Mixers
Buy Air Powered Mixers Items Including:
Lightnin general-purpose air-drive mixer
Low-cost, gear-drive air motor mixer; 8-170 rpm (YO-04681-00)
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00).
More Details
Air Powered Motor
Buy Air Powered Motor Items Including:
Air motor; 9000 rpm, 1200 cp (F 416-1 AIR)
Air motor; 900 rpm, 100,000 cp
Air motor; 900 rpm, 120,000 cp
Heavy-Duty TEFC Motor; 1-1/2 hp, 230/460 VAC, 60 Hz, 3 phase
Replacement motor, TEFC, 110 VAC, 60 Hz
More Details
Air Powered Peristaltic Pump
Buy Air Powered Peristaltic Pump Items Including:
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (S05B2P1TPNS000)
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (S07B1P1PPNS000.)
Sandpiper Air-Powered Double-Diaphragm PVDF Pump, PVDF, 23 GPM, Santop.
More Details
Air Powered Piston Pumps
Buy Air Powered Piston Pumps Items Including:
Series Q single head, air powered drive, 1800 spm (SPD).
More Details
Air Pressure
Buy Air Pressure Items Including:
Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 (960-115-000)
Hydronic Manometer (HM670) Water/Air Pressure Mtr
Brass vacuum/pressure gauge connector kit for all Air Cadet pumps
Eccentric assembly for all Air Cadet pumps.
More Details
Air Pressure Check Valve
Buy Air Pressure Check Valve Items Including:
Pressure Relief Valve Kit for use with the 68450-35
Graph-style aluminum flowmeter; withstand pressures to 1000 psi
Wette (YO-32206-02)
Graph-style aluminum flowmeter; withstand pressures to 1000 psi
Wette (YO-32206-04).
More Details
Air Pressure Gauge
Buy Air Pressure Gauge Items Including:
Air Filter Kit for Magnehelic Gauges
Fixed Snubber, 304/316 SS, 5000 psi, Air
Graph-style aluminum flowmeter with built-in psi gauge; withstand pres (YO-32206-52)
Graph-style aluminum flowmeter with built-in psi gauge; withstand pres (YO-32206-54).
More Details
Air Pressure Gauge 0 300 Lbs
Buy Air Pressure Gauge 0 300 Lbs Items Including:
Spriahelic 4 1/2" Direct Drive Pressure Gauge, 300 psi, 1/4" NPT(F) Ba
Differential Pressure Gauge with Membrane Sensing Element, 0-300 INWC;
SPAN Pressure Gauge, 0 to 300 psi (YO-68081-16)
SPAN Pressure Gauge, 0 to 300 psi (YO-68081-46).
More Details
Air Pressure Gauge Calibration
Buy Air Pressure Gauge Calibration Items Including:
Brass vacuum/pressure gauge connector kit for all Air Cadet pumps
Ashcroft Pressure Gauge Comparator, 30 psi
Ashcroft Pressure Gauge Comparator, 0-100 psi
Ashcroft Pressure Gauge Comparator, 1000 psi
Ashcroft Pressure Gauge Comparator, 10,000 psi.
More Details
Air Pressure Gauges 0 200 Psi
Buy Air Pressure Gauges 0 200 Psi Items Including:
Spriahelic 4 1/2" Direct Drive Pressure Gauge, 200 psi, 1/4" NPT(F) Ba
Digital Industrial Gauge, 200 psi, Battery Powered
Digital Industrial Gauge, 200 psi, Loop Powered
Brass vacuum/pressure gauge connector kit for all Air Cadet pumps.
More Details
Air Pressure Manometers
Buy Air Pressure Manometers Items Including:
Hydronic Manometer (HM670) Water/Air Pressure Mtr.
More Details
Air Pressure Meter
Buy Air Pressure Meter Items Including:
Hydronic Manometer (HM680) Water/Air Flow Meter
4005LKIT:Air Vlcty 0/5 Inwc Air Velocity M Kit 400/9000 Velocity Fpm W
40023KIT:Airvlcty 0/23 Inwc Air Velocity M Kit 400/19 200 Velocity Fpm.
More Details
Air Pressure Pump
Buy Air Pressure Pump Items Including:
Brass vacuum/pressure gauge connector kit for all Air Cadet pumps
Eccentric assembly for all Air Cadet pumps
Air Cadet Vacuum/Pressure Pump, OEM, 0.65 CFM, 20" HG, 115 VAC
Air Cadet Vacuum/Pressure Pump, Diaphragm, single head, 0.5 cfm, 230 V.
More Details
Air Pressure Regulators
Buy Air Pressure Regulators Items Including:
Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 (960-115-000)
Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 (960-116-000)
Marsh Bellofram Type 10 Pressure Regulator, 1/8" NPT(F), 2-25 psi.
More Details
Air Pressure Valves
Buy Air Pressure Valves Items Including:
Pressure Relief Valve Kit for use with the 68450-35
Graph-style aluminum flowmeter; withstand pressures to 1000 psi
Wette (YO-32206-02)
Graph-style aluminum flowmeter; withstand pressures to 1000 psi
Wette (YO-32206-04).
More Details
Air Probe
Buy Air Probe Items Including:
Wd-08500-75:T-Air Probe Type T Thermocouple Air Probe (071497A-T)
Thermocouple Air probe Type-K
Air probe for Model 4000 (90225-27)
Ap-k : Air Probe Type K (YO-08876-69)
Ap-j : Air Probe Type J (YO-08876-73)
More Details
Air Process Control Valves
Buy Air Process Control Valves Items Including:
Cole-Parmer 65-mm Direct Reading Flowmeter For Air; Brass Without Valv (YO-32011-16)
Cole-Parmer 65-mm Direct Reading Flowmeter For Air; Brass Without Valv (YO-32011-18).
More Details
Air Products
Buy Air Products Items Including:
Full-Facepiece Mask assembly for Supplied Air System
Nitrile diaphragm service kit for all Air Cadet pumps
Eccentric assembly for all Air Cadet pumps
Diaphragm shaft for all Air Cadet pumps
Test Products International Handheld Particle Counter.
More Details
Air Products Gas
Buy Air Products Gas Items Including:
Cal Gas, 5% CO2, 17 liters
MMC Card Reader; USB, for user-downloadable gas datalogger/detector
Honeywell Impact Pro Multigas Monitor Gas cartridge, O2, LE (2302BCS3OFAXO)
Honeywell Impact Pro Multigas Monitor Gas cartridge, O2, LE (2302BCS3OFDXO).
More Details
Air Products Regulators
Buy Air Products Regulators Items Including:
Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 (960-115-000)
Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 (960-116-000)
Marsh Bellofram Type 10 Pressure Regulator, 1/8" NPT(F), 2-25 psi.
More Details
Air Pump Bellows
Buy Air Pump Bellows Items Including:
Asti All (PFA-coated) Double-Bellows Pumps 10 LPM
Asti All (PFA-coated) Double-Bellows Pumps 30 LPM, 5 bar, 3/4" flare c.
More Details
Air Pump Compressors
Buy Air Pump Compressors Items Including:
Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4"
Mini Air Regulator, Max 45 SCFM, Pressure range of 0 - 140 PSIG, 1/8"
Compact Air Regulator, Max 86 SCFM, Pressure range of 0 - 140 PSIG, 1/.
More Details
Air Pump Diaphragm Kits
Buy Air Pump Diaphragm Kits Items Including:
Repair parts kit for high-purity air-operated double-diaphragm pumps 7 (K10-F)
Repair parts kit for high-purity air-operated double-diaphragm pumps 7 (K20-F)
Service Kit for 75320-90
Service kit for models 75300-02
Service kit for models 75300-12.
More Details
Air Pump for a Tube
Buy Air Pump for a Tube Items Including:
Pump and mix tube set, 39", Stainless Steel (F426S 41/38 39")
Pump and mix tube set, 39", Polypropylene (F426P 50/33 39")
High-viscosity tube set (F55GS50/2150/21-39)
High-head PP tube set for electric and air motors, 27"L.
More Details
Air Pump for Air Tanks
Buy Air Pump for Air Tanks Items Including:
Oilless Air Compressor with 20 Gallon Storage Tank, 3/4 hp, 115 VAC/60 (5HCD-11T-M550NGX)
Ultra-Quiet Oil-lubricated Air Compressor, 2.1 CFM (60 LPM), 6.3 Gallo
Ultra-Quiet Oil-lubricated Air Compressor, 4.2 CFM (120 LPM), 6.3 Gall (YO-07067-23).
More Details
Air Pump for Fluids
Buy Air Pump for Fluids Items Including:
Pump, Vacuum Pump, Rotary Vane, 1.5 CFM, 110VAC, 2 Stage Pump (UVP1.5-110) (YO-07164-00)
Pump, Vacuum Pump, Rotary Vane, 1.5 CFM, 220VAC, 2 Stage Pump (UVP1.5-220) (YO-07164-01).
More Details
Air Pump Inlet Outlet
Buy Air Pump Inlet Outlet Items Including:
Inlet/Outlet filter, 10 micron filter, 3.2 CFM, 1/8" NPT(F)
Inlet/Outlet filter, 10 micron filter, 31.8 CFM, 1/2" NPT(F).
More Details
70618
results
<<<Previous
56
57
58
59
60
61
62
63
64
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
45
46
47
48
49
50
51
52
53
54
55
56
57
58
59
60
61
62
63
64
65
66
67
68
69
70
71
72
73
74
80
90
100
110
120
130
140
150
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Air Powered Lab Stirrer to Air Pump Inlet Outlet
>
31125329-s-toc_2 31125329stoc2