Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 700
Eosin-Y Histology Stain
to
Epoxy Supplies
Image
Product Detail
More Detail
Eosin-Y Histology Stain
Buy Eosin-Y Histology Stain Items Including:
Eosin-Y, Phloxine, Four-1-pint Bottles (YO-48803-60)
Eosin Y Certified Powder, 50gm/ea
See MSDS for HAZ-MAT handling requi.
More Details
Ep Meter Myron L
Buy Ep Meter Myron L Items Including:
MYRON L EP analog conductivity/resistivity meter.
More Details
Epa Compliant
Buy Epa Compliant Items Including:
+GF+ Signet 4150 Turbidimeter, 0 to 1000 NTU, EPA compliant (YO-99500-02)
HF Scientific MicroTOL 2 Turbidimeter, 0 to 1000 NTU, EPA compliant
LaMotte 2020we Handheld Turbidity Meter, EPA compliant
Hach 2100Q Handheld Turbidity Meter, EPA compliant.
More Details
Epa Lab
Buy Epa Lab Items Including:
Turbidity Standard, 10 NTU, EPA, 60 mL bottle (1485)
Turbidity Standard, 100 NTU, EPA, 60 mL bottle (1486)
Wildco Surber Stream Bottom Sampler, 500 um (EPA) net (12-C52)
Wildco Kick Net 500 um (EPA) (425-K53)
Wildco Tri-Net 500 um (EPA) (425-T52).
More Details
Epa Sample Containers
Buy Epa Sample Containers Items Including:
Cole-Parmer Precleaned EPA Sample Jars, 60 mL
Cole-Parmer Precleaned EPA Sample Jars, 125 mL
Cole-Parmer Precleaned EPA Sample Jars, 250 mL
Cole-Parmer Precleaned EPA Sample Jars, 500 mL.
More Details
Epa Samplers
Buy Epa Samplers Items Including:
Wildco Surber Stream Bottom Sampler, 500 um (EPA) net (12-C52)
Wildco Hess Stream Bottom Sampler, 500 um (EPA) Net (16-C52)
Wildco D-frame Multifilment 500 um (EPA) Net (425-D52)
Wildco Kick Net 500 um (EPA) (425-K53).
More Details
Epa Test
Buy Epa Test Items Including:
LaMotte LTC3000we Benchtop Turbidity and Chlorine Meter, EPA compliant
LaMotte 2020we Handheld Turbidity Meter, EPA compliant
Hach 2100Q Handheld Turbidity Meter, EPA compliant
HF Scientific Micro 100 Benchtop Turbidity Meter, EPA compliant, 110 V.
More Details
Epa Turbidity
Buy Epa Turbidity Items Including:
Hach 2100Q Handheld Turbidity Meter, EPA compliant
LaMotte LTC3000we Benchtop Turbidity and Chlorine Meter, EPA compliant
LaMotte 2020we Handheld Turbidity Meter, EPA compliant
HF Scientific Micro 100 Benchtop Turbidity Meter, EPA compliant, 110 V.
More Details
Epa Vials
Buy Epa Vials Items Including:
Clear Precleaned EPA vials 20 mL case of 72 (YO-99535-05)
Clear Precleaned EPA vials, 40 mL, case of 72 (YO-99535-15)
Clear Precleaned EPA vials 60 mL case of 72 (YO-99535-25)
Heated Evaporator/Concentrator; single block, 8 EPA Vials (YO-28690-00).
More Details
Epa Water Testing
Buy Epa Water Testing Items Including:
LaMotte LTC3000we Benchtop Turbidity and Chlorine Meter, EPA compliant
LaMotte 2020we Handheld Turbidity Meter, EPA compliant
Hach 2100Q Handheld Turbidity Meter, EPA compliant
HF Scientific Micro 100 Benchtop Turbidity Meter, EPA compliant, 110 V.
More Details
Epa/Precleaned Vials
Buy Epa/Precleaned Vials Items Including:
Clear Precleaned EPA vials, 40 mL, case of 72 (YO-99535-15)
Amber Precleaned EPA vials, 40 mL, case of 72 (YO-99535-16)
Amber Precleaned EPA vials 60 mL, case of 72 (YO-99535-26)
Thermo Scientific I-CHEM pre-cleaned vial, 20 mL, case of 72.
More Details
Epdm Gaskets
Buy Epdm Gaskets Items Including:
Smart Gasket , 1/2" Tri-Clamp, 1 Port, EPDM, Each
Smart Gasket , 3/4" Tri-Clamp, 1 Port, EPDM, Each
Smart Gasket , 1 1/2" Tri-Clamp, 1 Port, EPDM, Each
Smart Gasket , 1 1/2" Tri-Clamp, 2 Port, EPDM, Each.
More Details
Epdm Hose
Buy Epdm Hose Items Including:
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 57 GPM, conti
Large-Hose Peristaltic Pump, Lubricant-Filled Pump Cavity, EPDM Hose, (3005-EF-PV-F2)
Large-Hose Peristaltic Pump, Lubricant-Filled Pump Cavity, EPDM Hose, (3005-EF-PV-J2).
More Details
Epdm Peristaltic Pump Hose
Buy Epdm Peristaltic Pump Hose Items Including:
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 57 GPM, conti
Large-Hose Peristaltic Pump, Lubricant-Filled Pump Cavity, EPDM Hose, (3005-EF-PV-F2)
Large-Hose Peristaltic Pump, Lubricant-Filled Pump Cavity, EPDM Hose, (3005-EF-PV-J2).
More Details
Epdm Peristaltic Pump Tubing Ft Myers
Buy Epdm Peristaltic Pump Tubing Ft Myers Items Including:
Masterflex® peroxide-cured silicone tubing, I/P® 89, 10 ft
(YO-96400-89)
Hose Replacement for 75800-6* pumps, EPDM, fiber braided, standard dut
Hose Replacement for 75800-5* pumps, EPDM, steel braided, standard dut
Replacement RJ12 cable, 3.5 ft..
More Details
Epdm Screen Gasket
Buy Epdm Screen Gasket Items Including:
Sock Screen Gasket, Epdm, 16-ss Mesh, 0.75 Length, 2 Clamp Size (SSG-E-S-200/16SS)
Sock Screen Gasket, Epdm, 16-ss Mesh, 0.75 Length, 3 Clamp Size (SSG-E-S-300/16SS)
Sock Screen Gasket, Epdm, 16-ss Mesh, 0.75 Length, 2 Clamp Size (SSG-E-L-200/16SS).
More Details
Epi Fluorescence Microscope
Buy Epi Fluorescence Microscope Items Including:
Meiji Epi-Fluorescent Microscope, Binocular
Meiji Epi-Fluorescent Microscope, Trinocular.
More Details
Epic Plastic Storage Bins
Buy Epic Plastic Storage Bins Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Epithelial Tissue Microscope Slides
Buy Epithelial Tissue Microscope Slides Items Including:
Cole-Parmer Tissue Flotation Work Center, 8"x8"x2" with slide dryer, 3 (YO-48579-00)
Cole-Parmer Tissue Flotation Work Center, 8"x8"x2" with slide dryer, 3 (YO-48579-02).
More Details
Epon Tissue Embedding and Processing
Buy Epon Tissue Embedding and Processing Items Including:
Tissue Processing Capsules, 8 mm D x 38 mm dia (6010)
Embedding Rings, Yellow (6001)
Embedding Rings, Pink (6002)
Embedding Rings, Green (6003)
Embedding Rings, Blue (6004)
More Details
Epoxy Body Orp Electrode
Buy Epoxy Body Orp Electrode Items Including:
Cole-Parmer pH electrode, epoxy-body, spear tip, double-junction, BNC
pH electrode, general purpose, combination, flat, sealed, epoxy body, (YO-05990-65)
Epoxy body, single junction ORP electrode
Cole-Parmer Removable Single-Junction ORP Electrode.
More Details
Epoxy Gel
Buy Epoxy Gel Items Including:
Accumet pH Electrode, Combination, General Purpose, Gel-filled, Epoxy
ORP electrode; single-junction, gel-filled, BNC connector
accumet accuCap, epoxy body, gel-filled
Thermo Scientific Orion pH/ATC Triode, epoxy body, sealed, single-junc (9107BN).
More Details
Epoxy Mixing
Buy Epoxy Mixing Items Including:
Tool box kit, includes 100 mixing sticks and assortment of 100 epoxies (YO-08778-30).
More Details
Epoxy Mixing Equipment
Buy Epoxy Mixing Equipment Items Including:
Tool box kit, includes 100 mixing sticks and assortment of 100 epoxies (YO-08778-30).
More Details
Epoxy Ph Probe
Buy Epoxy Ph Probe Items Including:
Cole-Parmer Epoxy Conductivity Probe for Traceable Meters K=1
Thermo Scientific Orion Epoxy Automatic Temperature Compensation Probe
Conductivity Probe, Epoxy, K=1.0 (YO-19601-07)
Conductivity probe, 4-cell, K = 0.1, Epoxy/platinum, 2-1/2 ft cable.
More Details
Epoxy Ph Probe Compatibility Chart
Buy Epoxy Ph Probe Compatibility Chart Items Including:
Cole-Parmer Epoxy Conductivity Probe for Traceable Meters K=1
Accumet pH Electrode, Combination, General Purpose, Refillable, Epoxy
Accumet pH Electrode, Combination, Flat, Refillable, Epoxy Body, BNC
Conductivity Probe, Epoxy, K=1.0 (YO-19601-07).
More Details
Epoxy Pump
Buy Epoxy Pump Items Including:
Sanitary Centrifugal Pump, Heavy-Duty, 50 GPM, 1 HP, White Epoxy Motor
Sanitary 3A Centrifugal Pump, 105 GPM, 1.5 HP, White Epoxy Motor, 3600
Sanitary 3A Centrifugal Pump, 148 GPM, 3 HP, White Epoxy Motor, 3600 R.
More Details
Epoxy Resin Tops
Buy Epoxy Resin Tops Items Including:
1 Drawer Wood Lab Table With Epoxy Resin Top 36in W X 30in H X 24in D (YO-41527-00)
2 Drawer Wood Lab Table With Epoxy Resin Top 48in W X 30in H X 24in D (YO-41527-02).
More Details
Epoxy Set
Buy Epoxy Set Items Including:
Epoxy,quick-set, 10/pk (YO-08778-02)
Epoxy, Quick-set, non sagging, 10/pk (YO-08778-04)
Epoxy, quick set, machinable, 10/pk (YO-08778-10).
More Details
Epoxy Supplies
Buy Epoxy Supplies Items Including:
Centrifuge tube rack for 15-mL centrifuge tubes (YO-06696-02)
Poxygrid Wire Test Tube Half Rack 36 10-13mm (YO-06739-10)
Poxygrid Wire Test Tube Half Rack 24 15-16mm (YO-06739-12)
Poxygrid Wire Test Tube Half Rack 20 18-20mm (YO-06739-14).
More Details
70618
results
<<<Previous
696
697
698
699
700
701
702
703
704
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
610
620
630
640
650
660
670
680
685
686
687
688
689
690
691
692
693
694
695
696
697
698
699
700
701
702
703
704
705
706
707
708
709
710
711
712
713
714
720
730
740
750
760
770
780
790
800
900
1,000
1,100
1,200
1,300
1,400
1,500
1,600
2,000
2,354
Eosin-Y Histology Stain to Epoxy Supplies
>
31125329-s-toc_2 31125329stoc2