Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 80
Amicon Centrifuge Filters
to
Ammonium Chloride Usp Grade
Image
Product Detail
More Detail
Amicon Centrifuge Filters
Buy Amicon Centrifuge Filters Items Including:
Millipore Amicon Centrifugal Filter Units, 10k NMWL, 24/pk, green
Millipore Amicon Centrifugal Filter Units, 100k NMWL, 24/pk, blue
Millipore Amicon Ultra Centrifugal Filters, 4ML 100K 24PK
Millipore Amicon Ultra Centrifugal Filters, 15ML 10K 24PK.
More Details
Amicon Ultra
Buy Amicon Ultra Items Including:
Millipore Amicon Ultra Centrifugal Filters, 4ML 100K 24PK
Millipore Amicon Ultra Centrifugal Filters, 15ML 10K 24PK
Millipore Amicon Ultra Centrifugal Filters, 15ML 100K 24PK
Millipore Amicon Ultra Centrifugal Filters, 0.5 mL, 3K NMWL; pack of 2.
More Details
Ammex Black Nitrile Gloves
Buy Ammex Black Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, X-large, Box of 50
Microflex MidKnight™ Black Nitrile Glove, small, 100/box
Microflex MidKnight™ Black Nitrile Glove, medium, 100/box
Microflex MidKnight™ Black Nitrile Glove, extra large, 100/box.
More Details
Ammonia Analysis
Buy Ammonia Analysis Items Including:
Sensidyne Gas Detection Tubes, Ammonia (MR)
Sensidyne Gas Detection Tubes, Ammonia (SEI)
RAE Systems gas detection tubes, ammonia, 5 to 100 ppm, 10/box
RAE Systems gas detection tubes, ammonia, 25 to 500 ppm, 10/box
CMS Chip, Ammonia, 2 to 50 ppm.
More Details
Ammonia and Water
Buy Ammonia and Water Items Including:
Ammonia High Range Reagent Set, 50 Tests (Rs5650)
Ammonia Nitrogen Reagent Set, 10 mL, 100 Tests (Rs5500)
Ammonia Nitrogen Reagent Set, Salicylate Method, 0-0.50 ppm, 100 Tests
Refill kit, Ammonia/nitrogen, Nesslerization test, for 00291-52, six-m.
More Details
Ammonia Electrode
Buy Ammonia Electrode Items Including:
ISE Standard solution, ammonia/ammonium, 0.1 M (YO-27503-00)
Ammonia fill solution, 0.1M NH4Cl, 125-mL bottle (YO-27503-71)
Replacement membranes for electrode 27502-00 (YO-27503-95)
Cole-Parmer ISE electrode, ammonia, BNC, with pre-membraned caps.
More Details
Ammonia Electrode Filling Solution
Buy Ammonia Electrode Filling Solution Items Including:
Ammonia fill solution, 0.1M NH4Cl, 125-mL bottle (YO-27503-71).
More Details
Ammonia Filter
Buy Ammonia Filter Items Including:
Ammonia filter cell (16 lb treated carbon media), for Filtermate exhau (3923402)
Ammonia & Amines Carbon Filter For 3-ft & 6-ft Xpert Systems (YO-09109-73)
Carbon Filter For Paramount Filtered Enclosures For Ammonia And Amines (YO-28613-80).
More Details
Ammonia Fume Hood
Buy Ammonia Fume Hood Items Including:
Single-Filter Ammonia Fume Adsorber, 115V (YO-09003-60)
Single-filter Ammonia Fume Adsorber, 230V (YO-09003-70)
Double-filter Ammonia Fume Adsorber, 115V (YO-09003-80)
Ammonia & Amines Carbon Filter For 2-ft Xpert Systems (YO-09109-72).
More Details
Ammonia Gas Analyzer
Buy Ammonia Gas Analyzer Items Including:
CMS Chip, Ammonia, 100 to 2000 ppm.
More Details
Ammonia Gas Detectors
Buy Ammonia Gas Detectors Items Including:
Portable Ammonia Detector, 0 to 100% LEL.
More Details
Ammonia Gasket Material Threaded Fitting
Buy Ammonia Gasket Material Threaded Fitting Items Including:
Push-to-connect threaded elbow adapters, polypropylene, 1/4" x 1/8" NP (YO-34007-62)
Push-to-connect threaded elbow adapters, polypropylene, 1/4" x 1/4" NP (YO-34007-63).
More Details
Ammonia Gauges Dual Scale
Buy Ammonia Gauges Dual Scale Items Including:
30 to 0" Hg Dual-Scale Gauge, 1 1/2" Dial
30" Hg to 100 psi Dual-Scale Gauge, 1 1/2" Dial
30 to 0" Hg and —100 to 0 kPa Dual-Scale Gauge, Bottom Connecti
30" Hg to 30 psi and —100 to 200 kPa Dual-Scale Gauge, Bottom C.
More Details
Ammonia Ion Electrode Calibration
Buy Ammonia Ion Electrode Calibration Items Including:
Filling solution f/ammonia ISE electrode (951202)
Ammonia standard, 1000 ppm ammonia as nitrogen, 475-mL bottle (YO-27503-93)
Ammonia fill solution, 0.1M NH4Cl, 125-mL bottle (YO-27503-71)
Replacement membranes for electrode 27502-00 (YO-27503-95).
More Details
Ammonia Ion Selective Electrode
Buy Ammonia Ion Selective Electrode Items Including:
Ammonia fill solution, 0.1M NH4Cl, 125-mL bottle (YO-27503-71)
Ammonia standard, 1000 ppm ammonia as nitrogen, 475-mL bottle (YO-27503-93)
Replacement membranes for electrode 27502-00 (YO-27503-95)
Filling solution f/ammonia ISE electrode (951202).
More Details
Ammonia Ise
Buy Ammonia Ise Items Including:
Filling Solution Ammonia ISE 60 mL bottle (951209)
Filling solution f/ammonia ISE electrode (951202)
Thermo Scientific Orion ISE ammonia/ammonium electrode with BNC connec (9512BNWP)
Cole-Parmer ISE electrode, ammonia, BNC, with pre-membraned caps.
More Details
Ammonia Ise Meters
Buy Ammonia Ise Meters Items Including:
Accumet XL250N pH/ISE Ammonia Benchtop Meter Kit
Ammonia ISE Replacement bodies with pre-installed membranes (951215)
Filling solution f/ammonia ISE electrode (951202)
Thermo Scientific Orion ISE electrode, Hi-Performance Ammonia, BNC (9512HPBNWP).
More Details
Ammonia Meters
Buy Ammonia Meters Items Including:
Thermo Scientific Orion Star A214 pH/ISE Ammonia Meter Kit
Ammonia Nitrogen Reagent Set, Salicylate Method, 0-0.50 ppm, 100 Tests
CHEMets Colorimetric Ammonia/nitrogen Test Kit
Refill kit, Ammonia/nitrogen, Nesslerization test, for 00291-52, six-m.
More Details
Ammonia Monitor
Buy Ammonia Monitor Items Including:
ChromAir Gas Monitoring Badges; Ammonia, box of 10.
Ammonia Tube, High Level, 50 Tests.
More Details
Ammonia Ph
Buy Ammonia Ph Items Including:
Accumet XL250N pH/ISE Ammonia Benchtop Meter Kit
Ammonia standard, 100 ppm ammonia as nitrogen, 475-mL bottle (YO-27503-94)
ISE Standard solution, ammonia/ammonium, 0.1 M (YO-27503-00)
Ammonia fill solution, 0.1M NH4Cl, 125-mL bottle (YO-27503-71).
More Details
Ammonia Pump
Buy Ammonia Pump Items Including:
Drager Tubes, Ammonia, 0.05 To 10%
Drager Tubes, Ammonia, 2.5 to 100 ppm.
More Details
Ammonia Selective Electrodes
Buy Ammonia Selective Electrodes Items Including:
Filling solution f/ammonia ISE electrode (951202)
Thermo Scientific Orion ISE ammonia/ammonium electrode with BNC connec (9512BNWP)
Cole-Parmer ISE electrode, ammonia, BNC
Ammonia standard, 1000 ppm ammonia as nitrogen, 475-mL bottle (YO-27503-93).
More Details
Ammonia Solution
Buy Ammonia Solution Items Including:
Filling solution f/ammonia ISE electrode (951202)
ISE Standard solution, ammonia/ammonium, 0.1 M (YO-27503-00)
Ammonia fill solution, 0.1M NH4Cl, 125-mL bottle (YO-27503-71)
Filling Solution Ammonia ISE 60 mL bottle (951209).
More Details
Ammonia Specific Ion Electrode
Buy Ammonia Specific Ion Electrode Items Including:
Filling solution f/ammonia ISE electrode (951202)
ISE electrode, ammonia, for controller 56105-00 (YO-27077-00)
Ammonia standard, 100 ppm ammonia as nitrogen, 475-mL bottle (YO-27503-94)
ISE Standard solution, ammonia/ammonium, 0.1 M (YO-27503-00).
More Details
Ammonium Acetate
Buy Ammonium Acetate Items Including:
ACETIC ACID AMMONIUM SALT 25G (YO-88360-56).
More Details
Ammonium Bromide
Buy Ammonium Bromide Items Including:
AMMONIUM BROMIDE PWD ACS 500G (YO-88053-78)
AMMONIUM BROMIDE PA 5G (YO-88213-84)
AMMONIUM BROMIDE PA 25G (YO-88213-85).
More Details
Ammonium Carbonate
Buy Ammonium Carbonate Items Including:
AMMONIUM CARBONATE HPLC 500 G (YO-88053-79).
More Details
Ammonium Chloride
Buy Ammonium Chloride Items Including:
AMMONIUM CHLORIDE USP 3KG (YO-88053-73)
AMMONIUM CHLORIDE USP 500G (YO-88053-74)
AMMONIUM CHLORIDE ACS 10KG (YO-88053-88).
More Details
Ammonium Chloride Acs 500g
Buy Ammonium Chloride Acs 500g Items Including:
AMMONIUM CHLORIDE USP 500G (YO-88053-74)
AMMONIUM CHLORIDE ACS 10KG (YO-88053-88)
AMMONIUM BROMIDE PWD ACS 500G (YO-88053-78).
More Details
Ammonium Chloride Usp Grade
Buy Ammonium Chloride Usp Grade Items Including:
AMMONIUM CHLORIDE USP 500G (YO-88053-74).
More Details
70618
results
<<<Previous
76
77
78
79
80
81
82
83
84
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
60
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
85
86
87
88
89
90
91
92
93
94
100
110
120
130
140
150
160
170
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Amicon Centrifuge Filters to Ammonium Chloride Usp Grade
>
31125329-s-toc_2 31125329stoc2