Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 800
Fluorinated Bottles
to
Foam Caps
Image
Product Detail
More Detail
Fluorinated Bottles
Buy Fluorinated Bottles Items Including:
Thermo Scientific Nalgene fluorinated wide-mouth bottle, 1 L
Thermo Scientific Nalgene fluorinated narrow-mouth bottle, 250 mL
Thermo Scientific Nalgene fluorinated narrow-mouth bottle, 1 L
Thermo Scientific Nalgene fluorinated wash bottle, 500 mL.
More Details
Fluorinated Carboy
Buy Fluorinated Carboy Items Including:
Thermo Scientific Nalgene Round Fluorinated Carboys with Grips, 20 L
Thermo Scientific Nalgene fluorinated carboy with spigot, 20 L
Thermo Scientific Nalgene fluorinated jerrican with handle, 10 L.
More Details
Fluorinated Ethylene Propylene
Buy Fluorinated Ethylene Propylene Items Including:
Thermo Scientific Nalgene Bottle drop-dispenser, FEP, 30ml, 1/pk
Thermo Scientific Nalgene Erlenmeyer flask of FEP with Tefzel ETFE scr (YO-06113-83).
More Details
Fluorinated Jerricans
Buy Fluorinated Jerricans Items Including:
Thermo Scientific Nalgene fluorinated jerrican with handle, 10 L
Thermo Scientific Nalgene fluorinated jerrican with handle, 20 L
Thermo Scientific Nalgene fluorinated carboy with spigot, 9 L.
More Details
Fluorine Ion Selective Electrode
Buy Fluorine Ion Selective Electrode Items Including:
Cole-Parmer Combination Ion Selective Electrodes, Bromide (Br)
Cole-Parmer Combination Ion Selective Electrodes, Copper (Cu+2)
Cole-Parmer Combination Ion Selective Electrodes, Cyanide (CN)
Cole-Parmer Combination Ion Selective Electrodes, Nitrate.
More Details
Fluorine Resistant Ph Probe
Buy Fluorine Resistant Ph Probe Items Including:
Eco pH Probe 10m
ECO Cond Probe 10M
ECO Cond Probe 4M
WTW Cond 3110 meter kit and TetraCon probe, 1-m cable
WTW Cond 3110 meter kit and TetraCon probe, 3-m cable
More Details
Fluorocarbon Coated Magnetic Stirring Rod
Buy Fluorocarbon Coated Magnetic Stirring Rod Items Including:
PTFE-coated lab spatula, with 1/2"L spooned end and 1 1/2"L flat round (D1069292)
PTFE-coated lab spatula, with 1 1/2"L flat tapered end and 1 5/8"L fla (D1069294)
Scienceware stainless steel vibrating spatula with PTFE coating.
More Details
Fluoroelastomer Tubing
Buy Fluoroelastomer Tubing Items Including:
Hubtron™ Pascal Fluoroelastomer Tubing, 1/16" x 1/8", 25' (YO-95227-00)
Hubtron™ Pascal Fluoroelastomer Tubing, 3/16" x 5/16", 25' (YO-95227-04)
Hubtron™ Advance Fluoroelastomer Tubing, 1/16" x 3/16", 25' (YO-95226-00).
More Details
Fluorometer Cuvette
Buy Fluorometer Cuvette Items Including:
Borosilicate Cuvettes for Turner Quantech™ Fluorometer, Pack of Twe.
More Details
Fluoropolyme Life Science
Buy Fluoropolyme Life Science Items Including:
Heat-on Multi-well Block System (fluoropolymer Coating) (YO-81228-00)
Heat-on 10ml Insert (fluoropolymer Coating) (YO-81228-20)
Heat-on 100ml Insert (fluoropolymer Coating) (YO-81228-35)
Heat-on 150ml Insert (fluoropolymer Coating) (YO-81228-40).
More Details
Fluoropolyme Rigid Tubing
Buy Fluoropolyme Rigid Tubing Items Including:
Corrugated FEP Tubing Compatible with Barbed Fittings, 1/4"ID x 9/32"O
Corrugated FEP Tubing Compatible with Barbed Fittings, 3/8"ID x 13/32"
Corrugated FEP Tubing Compatible with Compression Fittings, 21/64"ID x.
More Details
Fluoropolymer Coating
Buy Fluoropolymer Coating Items Including:
Heat-on Multi-well Block System (fluoropolymer Coating) (YO-81228-00)
Heat-on 10ml Insert (fluoropolymer Coating) (YO-81228-20)
Heat-on 100ml Insert (fluoropolymer Coating) (YO-81228-35)
Heat-on 150ml Insert (fluoropolymer Coating) (YO-81228-40).
More Details
Fluoropolymer Fittings
Buy Fluoropolymer Fittings Items Including:
Corrugated FEP Tubing Compatible with Compression Fittings, 7/32"ID x
Corrugated FEP Tubing Compatible with Compression Fittings, 21/64"ID x
Pipe fittings, Pipe plugs NPT(M), Kynar, 1/8", 5/8", 17/32"; 10/pack (YO-30706-21).
More Details
Fluoropolymer Pipe Plugs Npt(M)
Buy Fluoropolymer Pipe Plugs Npt(M) Items Including:
Threaded plug, PFA, 1/8" male NPT (YO-06546-37)
Threaded plug, PFA, 1/4" male NPT (YO-06546-38)
Threaded plug, PFA, 3/8" male NPT (YO-06546-39)
Threaded plug, PFA, 1/2" male NPT (YO-06546-40).
More Details
Fluoropolymer Specialty Tubing
Buy Fluoropolymer Specialty Tubing Items Including:
Corrugated FEP Tubing Compatible with Barbed Fittings, 1/4"ID x 9/32"O
Corrugated FEP Tubing Compatible with Barbed Fittings, 3/8"ID x 13/32"
Corrugated FEP Tubing Compatible with Compression Fittings, 21/64"ID x.
More Details
Fluoropolymer Tube Heating
Buy Fluoropolymer Tube Heating Items Including:
Heat-on Insert For 8x16mm Tubes (fluoropolymer Coating) (YO-81228-45)
Heat-on Insert For 4x20mm Tubes (fluoropolymer Coating) (YO-81228-50).
More Details
Fluoropolymer Tubing
Buy Fluoropolymer Tubing Items Including:
Tubing,pfa,1/8x1/4,250' (YO-95232-02)
Coiled FEP tubing, 1/8"ID x 1/4"OD, 2 ft long (YO-95821-00)
Coiled FEP tubing, 1/8"ID x 1/4"OD, 4 ft long (YO-95821-01)
Coiled FEP tubing, 1/4"ID x 3/8"OD, 2 ft long (YO-95821-02).
More Details
Fluoropolymer Tubing Life Science
Buy Fluoropolymer Tubing Life Science Items Including:
Heat-on Insert For 8x16mm Tubes (fluoropolymer Coating) (YO-81228-45)
Heat-on Insert For 4x20mm Tubes (fluoropolymer Coating) (YO-81228-50).
More Details
Fluoropolymer Tubing Pfa Tubing
Buy Fluoropolymer Tubing Pfa Tubing Items Including:
Tubing,pfa,1/8x1/4,250' (YO-95232-02)
Convoluted PFA tubing, 1/4"ID x 9/32"OD, 5 ft long (YO-06487-04)
Convoluted PFA tubing, 3/8"ID x 7/16"OD, 5 ft long (YO-06487-06).
More Details
Fluoropolymer Tubing Rigid Tubing
Buy Fluoropolymer Tubing Rigid Tubing Items Including:
Tubing,pfa,1/8x1/4,250' (YO-95232-02)
Coiled FEP tubing, 1/8"ID x 1/4"OD, 2 ft long (YO-95821-00)
Coiled FEP tubing, 1/8"ID x 1/4"OD, 4 ft long (YO-95821-01)
Coiled FEP tubing, 1/4"ID x 3/8"OD, 2 ft long (YO-95821-02).
More Details
Fluoropolymer Tubing Tubing, and Hose
Buy Fluoropolymer Tubing Tubing, and Hose Items Including:
Tubing,pfa,1/8x1/4,250' (YO-95232-02)
Coiled FEP tubing, 1/8"ID x 1/4"OD, 2 ft long (YO-95821-00)
Coiled FEP tubing, 1/8"ID x 1/4"OD, 4 ft long (YO-95821-01)
Coiled FEP tubing, 1/4"ID x 3/8"OD, 2 ft long (YO-95821-02).
More Details
Fluoropolymer Tubing Tubing, Fittings, and Hose
Buy Fluoropolymer Tubing Tubing, Fittings, and Hose Items Including:
Corrugated FEP Tubing Compatible with Barbed Fittings, 1/4"ID x 9/32"O
Corrugated FEP Tubing Compatible with Barbed Fittings, 3/8"ID x 13/32"
Corrugated FEP Tubing Compatible with Compression Fittings, 21/64"ID x.
More Details
Fluoroware 2 Way Valve 1 4
Buy Fluoroware 2 Way Valve 1 4 Items Including:
PTFE 2-Way Stopcock, 1/4" Orifice, 1/4" NPT(F) connections (YO-98515-82)
PP Three-Way, Two-Position Elliptic Valve, 1/4" NPT(F) connectors (223-PP-1)
PVDF Three-Way, Two-Position Elliptic Valve, 1/4" NPT(F) connectors (223-PV-1).
More Details
Flush Diaphragm Pressure Transducer
Buy Flush Diaphragm Pressure Transducer Items Including:
In-line Seal, 1 1/2" Process Conn, w/ 15 PSI Transmitter
In-line Seal, 1 1/2" Process Conn, w/ 30 PSI Transmitter
In-line Seal, 1 1/2" Process Conn, w/ 100 PSI Transmitter
In-line Seal, 2" Process Conn, w/ 15 PSI Transmitter.
More Details
Flush Mount Miniature Pressure Transducer
Buy Flush Mount Miniature Pressure Transducer Items Including:
8 IN Circular Chart Recorder - Pressure, 1 pen, Flush/back mount (YO-30018-13)
8 IN Circular Chart Recorder - Pressure, 2 pen, Flush/back mount (YO-30018-14)
8 IN Circular Chart Recorder - Pressure, 1 pen, Flush/back mount (YO-30018-23).
More Details
Flux High Pressure Pumps
Buy Flux High Pressure Pumps Items Including:
Masterflex L/S High-Pressure Pump System; 1 to 100 rpm, 90 to 260 VAC
Pump, High Precision - Ultra High Pressure Pumps, up to 18,000 psi, Co (F05SNXP1-SP1)
Ismatec™ high-pressure pump with Series Q pump heads, 0.016 to 19 m (MAKE-UP) (YO-78021-00).
More Details
Fm 120 Conductivity Meter
Buy Fm 120 Conductivity Meter Items Including:
Cole-Parmer Concentration Contoller Conductive Probe - P-120S Conducti.
More Details
Fmi Lab Pump Model Qb
Buy Fmi Lab Pump Model Qb Items Including:
Service kit for models 75300-02
Service kit for models 75300-12
Service kit for models 75300-04
Service kit for models 75300-14
Service kit for model 75320-60
More Details
Foam Brushes
Buy Foam Brushes Items Including:
Foam Laboratory Brush Set
Universal Foam Glassware Brush, 15"L
Foam Laboratory Washing Brush, 9"L
Foam Laboratory Washing Brush, 18"L
Foam-tipped Carboy Brush
More Details
Foam Caps
Buy Foam Caps Items Including:
Kimble 20mL Scintillation Vials, Glass, PP caps, foamed polyethylene l.
More Details
70618
results
<<<Previous
796
797
798
799
800
801
802
803
804
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
700
710
720
730
740
750
760
770
780
785
786
787
788
789
790
791
792
793
794
795
796
797
798
799
800
801
802
803
804
805
806
807
808
809
810
811
812
813
814
820
830
840
850
860
870
880
890
900
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
2,000
2,354
Fluorinated Bottles to Foam Caps
>
31125329-s-toc_2 31125329stoc2