Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 10
Single-Phase Odp Motors
to
Ultrasonic Processor Micro Cup Horn
Image
Product Detail
More Detail
Single-Phase Odp Motors
Buy Single-Phase Odp Motors Items Including:
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (G216)
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (K235)
Motor, General-Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/2 H (G232).
More Details
Three-Phase Odp Motors
Buy Three-Phase Odp Motors Items Including:
Motor, General-Purpose, Three-Phase, Open Drip-Proof, AC Motor, 1/3 HP (G240)
Motor, General-Purpose, Three-Phase, Open Drip-Proof, AC Motor, 1/3 HP (G201)
Motor, General-Purpose, Three-Phase, Open Drip-Proof, AC Motor, 1/2 HP (G241).
More Details
Three-Phase Stainless Steel
Buy Three-Phase Stainless Steel Items Including:
Stainless Steel NEMA Type C-face Rigid Base Three-phase Motor, 1/3 Hp, (N450)
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST562P13F)
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST564P13F).
More Details
Ac Motors Types
Buy Ac Motors Types Items Including:
Motor, General-Purpose, Three-Phase, Open Drip-Proof, AC Motor, 3 HP, (E784)
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV251N02SXB)
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV371N02YXB).
More Details
Washdown Duty Motors
Buy Washdown Duty Motors Items Including:
Washdown Duty NEMA Type C-face Rigid Base Three-phase Motor, 1/2 Hp, 1
Washdown Duty NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp,.
More Details
Three Phase Ac Motors
Buy Three Phase Ac Motors Items Including:
Motor, General-Purpose, Three-Phase, Open Drip-Proof, AC Motor, 1/3 HP (G240)
Motor, General-Purpose, Three-Phase, Open Drip-Proof, AC Motor, 1/3 HP (G201)
Motor, General-Purpose, Three-Phase, Open Drip-Proof, AC Motor, 1/2 HP (G241).
More Details
Single-Phase Washdown Motors
Buy Single-Phase Washdown Motors Items Including:
Washdown Duty NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp,
Washdown Duty NEMA Type C-face Rigid Base Single-phase Motor, 1/2 Hp,.
More Details
Cole-Parmer General Purpose Motor
Buy Cole-Parmer General Purpose Motor Items Including:
Motor, General Purpose, Single-Phase, ODP (Open-Drip-Proof) 1/4 HP, 56
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (G216)
Motor, General-Purpose, Three-Phase, Open Drip-Proof, AC Motor, 3 HP, (E784).
More Details
Three-Phase Washdown Motors
Buy Three-Phase Washdown Motors Items Including:
Washdown Duty NEMA Type C-face Rigid Base Three-phase Motor, 1/2 Hp, 1
Washdown Duty NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp,.
More Details
Motors Washdown
Buy Motors Washdown Items Including:
Washdown Duty NEMA Type C-face Rigid Base Three-phase Motor, 1/2 Hp, 1
Washdown Duty NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp,.
More Details
General Purpose Motors
Buy General Purpose Motors Items Including:
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, (56C34F5301/ D311)
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (G216)
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (K235).
More Details
Rigid Motor
Buy Rigid Motor Items Including:
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST562P13F)
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST564P13F)
Stainless Steel NEMA Type C-face Rigid Base Three-phase Motor, 1/3 Hp, (N450).
More Details
Cole-Parmer Ac Motors
Buy Cole-Parmer Ac Motors Items Including:
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (G216)
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST562P13F)
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST564P13F).
More Details
Cole Ac Motors
Buy Cole Ac Motors Items Including:
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (G216)
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (K235)
Motor, General-Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/2 H (G232).
More Details
General Purpose Totally Enclosed (Tefc)/Open D
Buy General Purpose Totally Enclosed (Tefc)/Open D Items Including:
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, (56C34F5301/ D311)
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, (56C17F5321/ G571)
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1 Hp, 36.
More Details
Rigid Steel
Buy Rigid Steel Items Including:
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST562P13F)
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST564P13F)
Stainless Steel NEMA Type C-face Rigid Base Three-phase Motor, 1/3 Hp, (N450).
More Details
Cole General Purpose Motor
Buy Cole General Purpose Motor Items Including:
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (G216)
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (K235)
Motor, General-Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/2 H (G232).
More Details
Hp 3 in 1
Buy Hp 3 in 1 Items Including:
General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1/3 Hp, 3
Lightnin Nettco S-Series Sanitary Mixer, gear-drive, 1-hp, 3-phase, 20 (YO-50323-76)
Lightnin Nettco S-Series Sanitary Mixer, gear-drive, 1-hp, 3-phase, 20 (YO-50323-84).
More Details
Three-Phase Explosion-Proof
Buy Three-Phase Explosion-Proof Items Including:
Explosion-proof Three-phase NEMA Type C-face Motor, 3/4 Hp, 208-230/46
Explosion-proof Single-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM,.
More Details
Ac Single Phase
Buy Ac Single Phase Items Including:
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (G216)
Motor, General Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/3 H (K235)
Motor, General-Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1/2 H (G232).
More Details
Phase Pumps
Buy Phase Pumps Items Including:
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, (56C34F5301/ D311)
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, (56C17F5321/ G571)
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1 Hp, 36.
More Details
Circular Paper
Buy Circular Paper Items Including:
Temperature recorder; 6" circular paper; 7-day; Fahrenheit scale; 115V (5016-9 F SCALE)
Temperature recorder; 6" circular paper; 7-day; Celsius scale; 115V (5016-9 C SCALE)
Chart Paper for 6" Circular Recorder; 24 hour, 0 to 100°F/°C.
More Details
Prominet Solenoid Metering Pump
Buy Prominet Solenoid Metering Pump Items Including:
Service Kit for Solenoid Metering Pump, 12 GPD
Service Kit for Solenoid Metering Pump, 21- 44 GPD
Service Kit for Solenoid Metering Pump, 94 GPD
Service Kit for Solenoid Metering Pump, 240 GPD
Service Kit for Solenoid Metering Pump, 500 GPD.
More Details
Pulsafeeder Solenoid Metering Pumps
Buy Pulsafeeder Solenoid Metering Pumps Items Including:
Service Kit for Solenoid Metering Pump, 6 GPD
Service Kit for Solenoid Metering Pump, 12 GPD
Service Kit for Solenoid Metering Pump, 240 GPD
Service Kit for Solenoid Metering Pump, 500 GPD
Pulsafeeder Manual Control Metering Pump, 44 gdp, 115 VAC.
More Details
Solenoid Metering Pumps
Buy Solenoid Metering Pumps Items Including:
Service Kit for Solenoid Metering Pump, 12 GPD
Service Kit for Solenoid Metering Pump, 21- 44 GPD
Service Kit for Solenoid Metering Pump, 94 GPD
Service Kit for Solenoid Metering Pump, 240 GPD
Service Kit for Solenoid Metering Pump, 500 GPD.
More Details
Miniature Solenoid Metering Pumps
Buy Miniature Solenoid Metering Pumps Items Including:
Service Kit for Solenoid Metering Pump, 12 GPD
Service Kit for Solenoid Metering Pump, 21- 44 GPD
Service Kit for Solenoid Metering Pump, 94 GPD
Service Kit for Solenoid Metering Pump, 240 GPD
Service Kit for Solenoid Metering Pump, 500 GPD.
More Details
Pulsafeeder Solenoid Metering Pump Service Kits
Buy Pulsafeeder Solenoid Metering Pump Service Kits Items Including:
Service Kit for Solenoid Metering Pump, 6 GPD
Service Kit for Solenoid Metering Pump, 12 GPD
Service Kit for Solenoid Metering Pump, 21- 44 GPD
Service Kit for Solenoid Metering Pump, 94 GPD
Service Kit for Solenoid Metering Pump, 240 GPD.
More Details
Pulsafeeder Accessories and Replacement Items
Buy Pulsafeeder Accessories and Replacement Items Items Including:
Service Kit for Solenoid Metering Pump, 6 GPD
Service Kit for Solenoid Metering Pump, 12 GPD
Service Kit for Solenoid Metering Pump, 21- 44 GPD
Service Kit for Solenoid Metering Pump, 94 GPD
Service Kit for Solenoid Metering Pump, 240 GPD.
More Details
Replacement Chuck
Buy Replacement Chuck Items Including:
Adjustable chuck for mixer 04341-10 (YO-04340-90)
Replacement Chuck 8.5mm
Replacement Chuck, 10.5mm
Replacement chuck for ServoDyne™ Mixers.
More Details
Ultrasonic Processor Micro Cup Horn
Buy Ultrasonic Processor Micro Cup Horn Items Including:
Micro Cup Horn (630-0447)
Qsonica Ultrasonic Cell Disruptor Cup Horn Stand
Cup horn (3") for use with ultrasonic processors
Cup horn (5.5") for use with ultrasonic processors
Dual Horn (without probes) for use with ultrasonic processors.
More Details
70618
results
<<<Previous
6
7
8
9
10
11
12
13
14
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Single-Phase Odp Motors to Ultrasonic Processor Micro Cup Horn
>
31125329-s-toc_2 31125329stoc2