Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 11
Glove Box Replacement Gloves
to
Temperature Controlled Test Chamber
Image
Product Detail
More Detail
Glove Box Replacement Gloves
Buy Glove Box Replacement Gloves Items Including:
Neoprene Gloves for Glove Boxes; size 8 1/2; 30"L (5005500)
Butyl Gloves; size 8 1/2; 30"L (50055-01)
Butyl Gloves; size 9 3/4; 30"L (50056-01)
Hypalon Gloves; size 8 1/2; 30"L
Replacement Inlet Or Outlet HEPA Filter (5225105).
More Details
Hypalon Gloves
Buy Hypalon Gloves Items Including:
Hypalon Gloves; size 8 1/2; 30"L
Hypalon Gloves; size 9 3/4; 30"L
North Hypalon drybox gloves, 15 mil, size 9-3/4, 1 pair.
More Details
Hypalon Accessories for all Glove Boxes
Buy Hypalon Accessories for all Glove Boxes Items Including:
Hypalon Gloves; size 8 1/2; 30"L.
More Details
Cole-Parmer Glove Box Replacement Gloves
Buy Cole-Parmer Glove Box Replacement Gloves Items Including:
Neoprene Gloves for Glove Boxes; size 8 1/2; 30"L (5005500)
Neoprene Gloves for Glove Boxes; size 9 3/4; 30"L (50056-00)
Butyl Gloves; size 8 1/2; 30"L (50055-01).
More Details
Hypalon Furniture & Fume Hoods
Buy Hypalon Furniture & Fume Hoods Items Including:
Hypalon Gloves; size 8 1/2; 30"L.
More Details
Glove Box Replacement
Buy Glove Box Replacement Items Including:
Neoprene Gloves for Glove Boxes; size 9 3/4; 30"L (50056-00)
Butyl Gloves; size 8 1/2; 30"L (50055-01)
Butyl Gloves; size 9 3/4; 30"L (50056-01)
Stainless steel support cart for glove box models 34752-20 and -22
Me (800-CART-ABD).
More Details
Hypalon Glove Box Replacement Gloves
Buy Hypalon Glove Box Replacement Gloves Items Including:
Hypalon Gloves; size 8 1/2; 30"L.
More Details
Hypalon Glove Boxes
Buy Hypalon Glove Boxes Items Including:
Hypalon Gloves; size 8 1/2; 30"L.
More Details
Accessories for all Glove Boxes
Buy Accessories for all Glove Boxes Items Including:
Cole-Parmer Economical Glove Box - Small
Latex Hands for Glove Boxes; size 7; 12/pack (5006200)
Iris diaphragm retrofit kit; order to fit your current glove box (800/IRIS/CONV)
Static Eliminator; removes static chages inside glove box (800-AS/SPI).
More Details
Cole-Parmer Accessories for all Glove Boxes
Buy Cole-Parmer Accessories for all Glove Boxes Items Including:
Neoprene Gloves for Glove Boxes; size 9 3/4; 30"L (50056-00)
Cole-Parmer Economical Glove Box - Small
Cole-Parmer Economical Glove Box - Large
Straight sleeves; 28"L with hands (5005800)
Accordian sleeves; 24"L with hands (5005900).
More Details
Cole Glove Box Replacement Gloves
Buy Cole Glove Box Replacement Gloves Items Including:
Neoprene Gloves for Glove Boxes; size 8 1/2; 30"L (5005500)
Neoprene Gloves for Glove Boxes; size 9 3/4; 30"L (50056-00)
Butyl Gloves; size 8 1/2; 30"L (50055-01).
More Details
Cole Accessories for all Glove Boxes
Buy Cole Accessories for all Glove Boxes Items Including:
Neoprene Gloves for Glove Boxes; size 9 3/4; 30"L (50056-00)
Cole-Parmer Economical Glove Box - Small
Cole-Parmer Economical Glove Box - Large
Straight sleeves; 28"L with hands (5005800)
Accordian sleeves; 24"L with hands (5005900).
More Details
Cole Gloves
Buy Cole Gloves Items Including:
Controlled atmosphere anaerobic glove box; 2 port; 110 VAC, 60 Hz (YO-34750-10)
Controlled atmosphere anaerobic glove box; 2 port; 220 VAC, 50 Hz (YO-34750-15)
Butyl Gloves; size 8 1/2; 30"L (50055-01)
Cole-Parmer Economical Glove Box - Small.
More Details
Brookfield Digital Viscometer Model Lvdve
Buy Brookfield Digital Viscometer Model Lvdve Items Including:
Replacement stand for viscometers (MODEL A)
Small sample adaptor for models 98945-00 and -05; 3 to 10 K cp
Small sample adapter for models 98945-10, -15, -20 and -25; 250 to 830
Replacement spindle for models 98945-00, -05, 50 to 100,000 cp (LV-2).
More Details
Brookfield Viscometer Spindles
Buy Brookfield Viscometer Spindles Items Including:
Replacement spindle for models 98945-10 and -15 (100 to 200,000 cp) an (RVHV-3)
Replacement spindle for models 98945-10 and -15 (200 to 400,000 cp) an (RVHV-4)
Replacement spindle for models 98945-10 and -15 (400 to 80,000 cp) and (RVHV-5).
More Details
Adjustable Water Temperature Control Unit
Buy Adjustable Water Temperature Control Unit Items Including:
Temperature control unit; 115 VAC, 60 Hz
Temperature control unit; 230 VAC, 50 Hz
Spindles for temp control unit and small sample adapters; 250 to 830 K (SC4-27)
Spindles for temp control unit and small sample adapters; 500 to 1.7 M (SC4-28).
More Details
Lab Stands
Buy Lab Stands Items Including:
Replacement stand for viscometers (MODEL A)
Mixer stand, 12" x 9-1/2" base with 5/8" dia x 24"L stainless steel ro
Bench stand for 59790-series hardness testers
Test stand for micro A durometers (OS-3)
Floor Stand for 81781-00.
More Details
Viscometer Small Sample
Buy Viscometer Small Sample Items Including:
Spindles for temp control unit and small sample adapters; 30 to 100 K (SC4-31)
Spindles for temp control unit and small sample adapters; 60 to 200 K (SC4-34)
Spindles for temp control unit and small sample adapters; 50 to 170 K (SC4-21).
More Details
Portable Viscometer Small Sample Size
Buy Portable Viscometer Small Sample Size Items Including:
Spindles for temp control unit and small sample adapters; 30 to 100 K (SC4-31)
Spindles for temp control unit and small sample adapters; 60 to 200 K (SC4-34)
Spindles for temp control unit and small sample adapters; 50 to 170 K (SC4-21).
More Details
Lab Oven with Digital Temp Control
Buy Lab Oven with Digital Temp Control Items Including:
Spindles for temp control unit and small sample adapters; 250 to 830 K (SC4-27)
Spindles for temp control unit and small sample adapters; 500 to 1.7 M (SC4-28)
Spindles for temp control unit and small sample adapters; 1.25 K to 4
(SC4-14).
More Details
Temperature Controller Small Appliances
Buy Temperature Controller Small Appliances Items Including:
Spindles for temp control unit and small sample adapters; 250 to 830 K (SC4-27)
Pump, Metering with 5 control methods, 0.66gph, PVDF, Fkm Front Contro (96470133)
Pump, Metering with 5 control methods, 4.88gph, PVDF, Fkm Front Contro (96470352).
More Details
Small Humidity Test Chamber
Buy Small Humidity Test Chamber Items Including:
Replacement sample chamber for spindle 98945-88 (SC4-6R)
Replacement sample chamber for spindle 98945-89
Replacement sample chamber for spindle 98945-90 (SC4-8R).
More Details
Brookfield Viscometer Temperature Control
Buy Brookfield Viscometer Temperature Control Items Including:
Temperature control unit; 115 VAC, 60 Hz
Temperature control unit; 230 VAC, 50 Hz
Spindles for temp control unit and small sample adapters; 250 to 830 K (SC4-27)
Spindles for temp control unit and small sample adapters; 500 to 1.7 M (SC4-28).
More Details
Replacement Spindles for Brookfield Rotational Viscometers
Buy Replacement Spindles for Brookfield Rotational Viscometers Items Including:
Replacement spindle for models 98945-10 and -15 (100 to 200,000 cp) an (RVHV-3)
Replacement spindle for models 98945-10 and -15 (200 to 400,000 cp) an (RVHV-4)
Replacement spindle for models 98945-10 and -15 (400 to 80,000 cp) and (RVHV-5).
More Details
Small Sample Adapters
Buy Small Sample Adapters Items Including:
Spindles for temp control unit and small sample adapters; 250 to 830 K (SC4-27)
Spindles for temp control unit and small sample adapters; 500 to 1.7 M (SC4-28)
Spindles for temp control unit and small sample adapters; 1.25 K to 4
(SC4-14).
More Details
Vent Unit and Temperature Control
Buy Vent Unit and Temperature Control Items Including:
Temperature control unit; 115 VAC, 60 Hz
Temperature control unit; 230 VAC, 50 Hz
Spindles for temp control unit and small sample adapters; 250 to 830 K (SC4-27)
Spindles for temp control unit and small sample adapters; 500 to 1.7 M (SC4-28).
More Details
Rotational Viscometer Spindle Selection
Buy Rotational Viscometer Spindle Selection Items Including:
Spindles for temp control unit and small sample adapters; 250 to 830 K (SC4-27)
Spindles for temp control unit and small sample adapters; 500 to 1.7 M (SC4-28)
Spindles for temp control unit and small sample adapters; 1.25 K to 4
(SC4-14).
More Details
Accessories for Brookfield Rotational Viscometers
Buy Accessories for Brookfield Rotational Viscometers Items Including:
Replacement spindle for models 98945-10 and -15 (100 to 200,000 cp) an (RVHV-3)
Replacement spindle for models 98945-10 and -15 (200 to 400,000 cp) an (RVHV-4)
Replacement spindle for models 98945-10 and -15 (400 to 80,000 cp) and (RVHV-5).
More Details
Viscometer Spindles
Buy Viscometer Spindles Items Including:
Spindles for temp control unit and small sample adapters; 250 to 830 K (SC4-27)
Spindles for temp control unit and small sample adapters; 500 to 1.7 M (SC4-28)
Spindles for temp control unit and small sample adapters; 1.25 K to 4
(SC4-14).
More Details
Temperature Controlled Test Chamber
Buy Temperature Controlled Test Chamber Items Including:
Temperature Control System for Spectrophotometers, 15 to 40°C
Combination Temperature Controller and Sipper Systems for Spectrophoto
Replacement sample chamber for spindle 98945-88 (SC4-6R)
Replacement sample chamber for spindle 98945-90 (SC4-8R).
More Details
70618
results
<<<Previous
7
8
9
10
11
12
13
14
15
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Glove Box Replacement Gloves to Temperature Controlled Test Chamber
>
31125329-s-toc_2 31125329stoc2