Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 14
Replacement Threaded Tips
to
Compressor Hose Fittings
Image
Product Detail
More Detail
Replacement Threaded Tips
Buy Replacement Threaded Tips Items Including:
Replacement Threaded Tip 1/2" (flat) (YO-04710-40)
Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1" di
Ultrasonic Cell Disruptor Replacement Tips, 1/2" dia
Ultrasonic Cell Disruptor Replacement Tips, 3/4" dia.
More Details
Cole-Parmer Ultrasonic Processor Cup Horn
Buy Cole-Parmer Ultrasonic Processor Cup Horn Items Including:
Cup horn; 2-1/2" dia (YO-04710-38)
Micro Cup Horn (630-0447).
More Details
Product Type Continuous Flow Cell
Buy Product Type Continuous Flow Cell Items Including:
QSonica Sonicator Continuous Flow Cell.
More Details
Cole-Parmer Ultrasonic Processor Sound-Abating Enclosure
Buy Cole-Parmer Ultrasonic Processor Sound-Abating Enclosure Items Including:
Ultrasonic Processor Sound-Abating enclosure; 30"H.
More Details
Ultrasonic Booster
Buy Ultrasonic Booster Items Including:
Booster for use to process difficult samples (YO-04710-84)
Booster (2:1 gain) for use with ultrasonic processors.
More Details
Ultrasonic Processor Sound-Abating Enclosure
Buy Ultrasonic Processor Sound-Abating Enclosure Items Including:
Ultrasonic Processor Sound-Abating enclosure; 30"H.
More Details
Horn Cup
Buy Horn Cup Items Including:
Qsonica Ultrasonic Cell Disruptor Cup Horn Stand
Ultrasonic Cell Disruptor Cup Horn, 5-1/2" I.D.
Micro Cup Horn (630-0447)
Cup horn (3") for use with ultrasonic processors
Cup horn (5.5") for use with ultrasonic processors.
More Details
Continuous Flow Cell
Buy Continuous Flow Cell Items Including:
QSonica Sonicator Continuous Flow Cell.
More Details
Cole-Parmer Tapered Microtips
Buy Cole-Parmer Tapered Microtips Items Including:
Tapered microtips; 1/8" (YO-04710-42)
Tapered microtips; 3/16" (YO-04710-44).
More Details
Cole-Parmer Replacement Threaded Tips
Buy Cole-Parmer Replacement Threaded Tips Items Including:
Replacement Threaded Tip 1/2" (flat) (YO-04710-40)
Replacement Threaded Tip 3/4" (flat) (YO-04710-41)
Replacement Threaded Tip 1 in tip (flat) (YO-04710-47)
Threaded probes with replaceable 1/2 (YO-04710-31).
More Details
Tapered Microtips
Buy Tapered Microtips Items Including:
Tapered microtips; 1/8" (YO-04710-42)
Tapered microtips; 3/16" (YO-04710-44).
More Details
Cole-Parmer Multi-Element Probes
Buy Cole-Parmer Multi-Element Probes Items Including:
4-element probe (630-0559)
8-element probe (630-0586)
24-element probe (630-0579).
More Details
Stainless Temperature Probe
Buy Stainless Temperature Probe Items Including:
Temperature probe, stainless steel (YO-04711-50)
High Accuracy Dual-Input Thermometer with one stainless-steel probe
High Accuracy Dual-Input Thermometer with two stainless-steel probe
T-Bar stem thermometer with a 4.9" stainless steel probe.
More Details
Multi-Element Probes
Buy Multi-Element Probes Items Including:
4-element probe (630-0559)
8-element probe (630-0586)
24-element probe (630-0579).
More Details
Cole-Parmer Cup Horn
Buy Cole-Parmer Cup Horn Items Including:
Cup horn; 2-1/2" dia (YO-04710-38)
Micro Cup Horn (630-0447).
More Details
3 Watts
Buy 3 Watts Items Including:
Threaded probes with replaceable 3/4 (YO-04710-33)
Tapered microtips; 3/16" (YO-04710-44)
Extender; 3/4" tip (YO-04710-81)
Probe, 3-mm (1/8 (630-0422)
Immersion heater, loop, 3-1/8" dia
More Details
Cole Tips
Buy Cole Tips Items Including:
Replacement Threaded Tip 1 in tip (flat) (YO-04710-47)
Extender; 1" tip (YO-04710-82)
Cole-Parmer In-Line pH Electrode with Tuff-Tip, 100Ω ATC Ele (YO-05995-02)
Cole-Parmer In-Line pH Electrode with Tuff-Tip, Double-Junction, No (YO-05995-10).
More Details
Vacuum Regulator
Buy Vacuum Regulator Items Including:
Vacuum regulator, 0 to 29" Hg, 2.47cfm (AI3600003)
Vacuum Regulator, 0 to 30" Hg, 2 cfm Max Capacity (AJ675)
Pressure Regulator, 0 to 125 psig, 20 cfm Capacity (R262)
Pressure Regulator, 0 to 100 psig, 2 cfm Capacity (AJ674).
More Details
150 Psi Micro Water Pump
Buy 150 Psi Micro Water Pump Items Including:
Mini Lubricator with bowl guard, Max 32 SCFM, Max Pressure of 150 PSI,
Compact Air Filter, 5 micron filter, Max 73 SCFM, Max Pressure 150 PSI
Thermo Scientific Nalgene Aspirator pump, 7.5 psi minimum water pressu.
More Details
Compressor Filters
Buy Compressor Filters Items Including:
Mini Air Regulator, Max 47 SCFM, 5 Micron Filter, Pressure range of 0
Combo Filter/Regulator and Lubricator, Max 44 SCFM, Max Pressure of 15
Inlet filter/silencer assembly, 1/4" NPT(M), 4 cfm, Paper
Compressed-air filter, 6 cfm (8A02N-OB2-BX).
More Details
Aro Pressure Regulator
Buy Aro Pressure Regulator Items Including:
Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4"
Mini Air Regulator, Max 45 SCFM, Pressure range of 0 - 140 PSIG, 1/8"
Compact Air Regulator, Max 86 SCFM, Pressure range of 0 - 140 PSIG, 1/.
More Details
Air Compressor Inlet Filter
Buy Air Compressor Inlet Filter Items Including:
Inlet filter/silencer assembly, 1/2" NPT(M), 12 cfm, Paper
Inlet/Outlet filter, 10 micron filter, 3.2 CFM, 1/8" NPT(F)
Inlet/Outlet filter, 10 micron filter, 31.8 CFM, 1/2" NPT(F).
More Details
Falcon Air Compressor Pumps
Buy Falcon Air Compressor Pumps Items Including:
Air Compressor hose, 1/4 in x 25' PU braid air hose with swivel fittin (AA1061-blue)
Oilless Air Compressor, Piston compressor pump, 1.3 cfm, 115 VAC (1HAB-10-M100X)
Oilless Air Compressor, Piston compressor pump, 1.3 cfm, 220 VAC (1HAE-10-M104X).
More Details
Air Line Quick Disconnect Dust Cover
Buy Air Line Quick Disconnect Dust Cover Items Including:
Quick Disconnect Air Coupler, 1/4 in NPT(F) (MS-1/4F)
Quick Disconnect Air Coupler, 3/8 in NPT(F) (MS-3/8F)
Quick Disconnect Air Coupler, 1/4 in NPT(M) (MS-1/4M)
Quick Disconnect Air Coupler, 3/8 in NPT(M) (MS-3/8M).
More Details
Air Compressor Silencer
Buy Air Compressor Silencer Items Including:
Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 55 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 155 SCFM
Inlet filter/silencer assembly, 1/2" NPT(M), 12 cfm, Paper.
More Details
Air Compressor Brass Fittings
Buy Air Compressor Brass Fittings Items Including:
Flowmeter Kit, Brass Fittings; 1.2 LPM Air (ACBKT2GL)
Flowmeter Kit, Brass Fittings; 1.2 LPM Air With Valve (ACBKT2GL-V)
Flowmeter Kit, Brass Fittings; 2.5 LPM Air (ACBKT2ST)
Flowmeter Kit, Brass Fittings; 2.5 LPM Air With Valve (ACBKT2ST-V).
More Details
Vacuum Pump Silencer
Buy Vacuum Pump Silencer Items Including:
Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 55 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 155 SCFM
Replacement silencer for vacuum pumps 78165-10 and -20.
More Details
Pump Silencer
Buy Pump Silencer Items Including:
Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 55 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 155 SCFM
Inlet filter/silencer assembly, 1/2" NPT(M), 12 cfm, Paper.
More Details
Air Compressor Flow Meter
Buy Air Compressor Flow Meter Items Including:
Refrigeration Dryer for Compressors, 10 SCFM nominal flow, 115 VAC, 60
Refrigeration Dryer for Compressors, 15 SCFM nominal flow, 115 VAC, 60
General Tools and Instruments Flow Watch for Air/Water Speed & Tempera.
More Details
Compressor Hose Fittings
Buy Compressor Hose Fittings Items Including:
Air Compressor hose, 1/4 in x 25' PU braid air hose with swivel fittin (AA1061-blue)
Aluminum hose barb adapter, NW 16 to 10 mm (662511)
Adapter,1/2" Hose Barb (C105-14-645)
Adapter, NW 16 to 3/4" hose barb , 316L SS (C105-04-105).
More Details
70618
results
<<<Previous
10
11
12
13
14
15
16
17
18
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Replacement Threaded Tips to Compressor Hose Fittings
>
31125329-s-toc_2 31125329stoc2