Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 18
Air Compressors and Accessories
to
Vacuum Air Compressor
Image
Product Detail
More Detail
Air Compressors and Accessories
Buy Air Compressors and Accessories Items Including:
Air Compressor hose, 1/4 in x 25' PU braid air hose with swivel fittin (AA1061-blue)
Air Compressor hose, 1/4 in x 50' PU braid air hose with swivel fittin (AA1062-blue)
Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4".
More Details
Refrigeration Dryers
Buy Refrigeration Dryers Items Including:
Refrigeration Dryer for Compressors, 10 SCFM nominal flow, 115 VAC, 60
Refrigeration Dryer for Compressors, 15 SCFM nominal flow, 115 VAC, 60
Refrigeration Dryer for Compressors, 25 SCFM nominal flow, 115 VAC, 60.
More Details
Air Compressor Lines
Buy Air Compressor Lines Items Including:
Air Tool, Straight Line Sander, 16 in x 2-1/2 in pad, 2,500 rpm, 1/4 i (AT-7019)
Quick Disconnect Air Coupler, 1/4 in NPT(F) (MS-1/4F)
Quick Disconnect Air Coupler, 3/8 in NPT(F) (MS-3/8F)
Quick Disconnect Air Coupler, 1/4 in NPT(M) (MS-1/4M).
More Details
Air Filter Accessories
Buy Air Filter Accessories Items Including:
Mini Coalescing Air Filter, 0.3 micron coalescing filter, Max 49 SCFM,
Compac Coalescing Air Filter, 0.3 micron coalescing filter, Max 73 SCF
High-efficiency Filter for use with Ductless Air Filter System (YO-33703-41).
More Details
Air Compressor Use
Buy Air Compressor Use Items Including:
Pressure Relief Valve Kit for use with the 68450-35.
More Details
Fluke Air Compressors
Buy Fluke Air Compressors Items Including:
Pressure Relief Valve Kit for use with the 68450-35.
More Details
Cole-Parmer Air Preparation Systems
Buy Cole-Parmer Air Preparation Systems Items Including:
Coalescing Filter/Regulator, 1/4" NPT(F) connection port.
More Details
Air Filters for Air Compressors
Buy Air Filters for Air Compressors Items Including:
Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4"
Mini Air Regulator, Max 45 SCFM, Pressure range of 0 - 140 PSIG, 1/8"
1/4" NPT(F) Filter Regulator
Inlet filter/silencer assembly, 1/2" NPT(M), 12 cfm, Paper.
More Details
Cole Quick-Disconnect Couplings for Air Lines
Buy Cole Quick-Disconnect Couplings for Air Lines Items Including:
Quick Disconnect Air Coupler, 1/4 in NPT(F) (MS-1/4F)
Quick Disconnect Air Coupler, 3/8 in NPT(F) (MS-3/8F)
Quick Disconnect Air Coupler, 1/4 in NPT(M) (MS-1/4M)
Quick Disconnect Air Coupler, 3/8 in NPT(M) (MS-3/8M).
More Details
150 Psi Air Compressor
Buy 150 Psi Air Compressor Items Including:
Mini Lubricator with bowl guard, Max 32 SCFM, Max Pressure of 150 PSI,.
More Details
Compressor Regulators
Buy Compressor Regulators Items Including:
Pressure Regulator, 0 to 125 psig, 20 cfm Capacity (R262)
Vacuum regulator, 0 to 29" Hg, 2.47cfm (AI3600003)
Vacuum Regulator, 0 to 30" Hg, 2 cfm Max Capacity (AJ675)
Compact Air Regulator, Max 86 SCFM, Pressure range of 0 - 140 PSIG, 1/.
More Details
Cole Filter Regulators and Lubricators
Buy Cole Filter Regulators and Lubricators Items Including:
1/4" NPT(F) Filter Regulator
1/4" NPT(F) Filter Regulator Lubricator
3/8" NPT(F) Filter Regulator.
More Details
Pump Hose Kit
Buy Pump Hose Kit Items Including:
Pump, Utility Accessory Hose kit consisting of 10' garden hose, SST sp.
More Details
Filter Regulators
Buy Filter Regulators Items Including:
1/4" NPT(F) Filter Regulator Lubricator
Coalescing Filter/Regulator, 1/4" NPT(F) connection port
Combo Filter/Regulator and Lubricator, Max 44 SCFM, Max Pressure of 15
Mini Air Regulator, Max 47 SCFM, 5 Micron Filter, Pressure range of 0.
More Details
Cole-Parmer Absorptive Silencers
Buy Cole-Parmer Absorptive Silencers Items Including:
Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 55 SCFM.
More Details
Compressor Plugs
Buy Compressor Plugs Items Including:
Quick Disconnect Air Plug, 1/4 in NPT(F) (MP-1/4F)
Quick Disconnect Air Plug, 3/8 in NPT(F) (MP-3/8F).
More Details
Pumps and Compressors
Buy Pumps and Compressors Items Including:
Compressor with 2 gallon tank, 1.4 cfm, 115 VAC (160493)
Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 55 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 155 SCFM.
More Details
Air Quick
Buy Air Quick Items Including:
1/4" Quick-Disconnect Brass Coupler with Shut-Off Valve, 1/4" NPT(F) (A184)
Quick Disconnect Air Coupler, 1/4 in NPT(F) (MS-1/4F)
Quick Disconnect Air Coupler, 3/8 in NPT(F) (MS-3/8F)
Quick Disconnect Air Coupler, 1/4 in NPT(M) (MS-1/4M).
More Details
Pneumatic and Hydraulic
Buy Pneumatic and Hydraulic Items Including:
Hydraulic test hose kit for use with 68450-35
Pressure Relief Valve Kit for use with the 68450-35.
More Details
Cole-Parmer Refrigeration Dryers
Buy Cole-Parmer Refrigeration Dryers Items Including:
Refrigeration Dryer for Compressors, 10 SCFM nominal flow, 115 VAC, 60
Refrigeration Dryer for Compressors, 15 SCFM nominal flow, 115 VAC, 60
Refrigeration Dryer for Compressors, 25 SCFM nominal flow, 115 VAC, 60.
More Details
Size Air Filter
Buy Size Air Filter Items Including:
Mid Size Air Filter, 5 micron filter, Max 156 SCFM, Max Pressure 150 P
Mid Size Air Filter, 5 micron filter, Max 197 SCFM, Max Pressure 150 P
Replacement Compressed-air filter; 6.0cfm; port size; 1/4" NPT(F); 5/b (5/050-05-BX).
More Details
Filter Lubricator
Buy Filter Lubricator Items Including:
Combo Filter/Regulator and Lubricator, Max 44 SCFM, Max Pressure of 15
Combo Filter and Regulator and Lubricator, Max 61 SCFM, Max Pressure o
Combo Filter/Regulator and Lubricator, Max 71 SCFM, Max Pressure of 15.
More Details
Air Pump Compressors
Buy Air Pump Compressors Items Including:
Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4"
Mini Air Regulator, Max 45 SCFM, Pressure range of 0 - 140 PSIG, 1/8"
Compact Air Regulator, Max 86 SCFM, Pressure range of 0 - 140 PSIG, 1/.
More Details
Compressor and Vacuum
Buy Compressor and Vacuum Items Including:
Absorptive Silencer for Compressors or Vacuum Pumps, 155 SCFM
Rotary vane vacuum pump for corrosive gases, 0.9 cfm, 115 VAC
Rotary vane vacuum pump for corrosive gases, 0.9 cfm, 230 VAC
Rotary vane vacuum pump for corrosive gases, 5.6 cfm, 115 VAC.
More Details
Absorptive Silencers
Buy Absorptive Silencers Items Including:
Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 55 SCFM.
More Details
Compressor Couplers
Buy Compressor Couplers Items Including:
Quick Disconnect Air Coupler, 1/4 in NPT(F) (MS-1/4F)
Quick Disconnect Air Coupler, 3/8 in NPT(F) (MS-3/8F)
Quick Disconnect Air Coupler, 1/4 in NPT(M) (MS-1/4M)
Quick Disconnect Air Coupler, 3/8 in NPT(M) (MS-3/8M).
More Details
Air Compressor Valves
Buy Air Compressor Valves Items Including:
1/4" Quick-Disconnect Brass Coupler with Shut-Off Valve, 1/4" NPT(F) (A184).
More Details
Coalescing Filters
Buy Coalescing Filters Items Including:
Coalescing Filter/Regulator, 1/4" NPT(F) connection port
Mini Coalescing Air Filter, 0.3 micron coalescing filter, Max 49 SCFM,
Compac Coalescing Air Filter, 0.3 micron coalescing filter, Max 73 SCF.
More Details
Air Filters Air
Buy Air Filters Air Items Including:
Mini Coalescing Air Filter, 0.3 micron coalescing filter, Max 49 SCFM,
Compac Coalescing Air Filter, 0.3 micron coalescing filter, Max 73 SCF
Compact Air Filter, 5 micron filter, Max 73 SCFM, Max Pressure 150 PSI
1/4" NPT(F) Filter Regulator.
More Details
Vacuum Air Compressor
Buy Vacuum Air Compressor Items Including:
Direct-drive rotary vane vacuum pump, dual mode, 5.9 cfm, 115/230 VAC
Vacuum regulator, 0 to 29" Hg, 2.47cfm (AI3600003)
Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM
Absorptive Silencer for Compressors or Vacuum Pumps, 55 SCFM.
More Details
70618
results
<<<Previous
14
15
16
17
18
19
20
21
22
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Air Compressors and Accessories to Vacuum Air Compressor
>
31125329-s-toc_2 31125329stoc2