Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 20
Cole-Parmer Accessories for Dredges
to
Fast-Freeze Lab Equipment
Image
Product Detail
More Detail
Cole-Parmer Accessories for Dredges
Buy Cole-Parmer Accessories for Dredges Items Including:
Winch, 1000 lb maximum weight limit (66-C10)
Winch mounting plate (66-C52)
Split barrel messenger, 11 oz (45-B10)
Solid barrel messenger, 8 oz (46-D80).
More Details
Accessories for Dredges
Buy Accessories for Dredges Items Including:
Winch, 1000 lb maximum weight limit (66-C10)
Winch mounting plate (66-C52)
Split barrel messenger, 11 oz (45-B10)
Solid barrel messenger, 8 oz (46-D80).
More Details
Cole Tapered Nose Messengers
Buy Cole Tapered Nose Messengers Items Including:
Split barrel messenger, 11 oz (45-B10).
More Details
Cole-Parmer Winch
Buy Cole-Parmer Winch Items Including:
Winch, 1000 lb maximum weight limit (66-C10)
Winch mounting plate (66-C52).
More Details
Cole Accessories for Dredges
Buy Cole Accessories for Dredges Items Including:
Winch, 1000 lb maximum weight limit (66-C10)
Winch mounting plate (66-C52)
Split barrel messenger, 11 oz (45-B10)
Solid barrel messenger, 8 oz (46-D80).
More Details
Water Barrels
Buy Water Barrels Items Including:
Split barrel messenger, 11 oz (45-B10).
More Details
Cole Tapered Nose Me
Buy Cole Tapered Nose Me Items Including:
Split barrel messenger, 11 oz (45-B10).
More Details
Tapered Nose Me
Buy Tapered Nose Me Items Including:
Split barrel messenger, 11 oz (45-B10).
More Details
Cole Winch
Buy Cole Winch Items Including:
Winch, 1000 lb maximum weight limit (66-C10)
Winch mounting plate (66-C52).
More Details
16 Oz Plastic Bottles
Buy 16 Oz Plastic Bottles Items Including:
Honeywell Personal Eyewash Wall Stations, 16 oz refill bottle
Thermo Scientific Nalgene polypropylene narrow-mouth bottle, 16 oz
Cole-Parmer Bottle, Amber Wide Mouth Packers, 16 oz, 12/case
Wheaton Safety bottles with PTFE-lined caps; 16 oz, clear.
More Details
Accessories for Eyewashes
Buy Accessories for Eyewashes Items Including:
Honeywell Personal Eyewash Wall Stations, 16 oz refill bottle
Bacteria-inhibiting eyewash preservative (32-001100-0000)
Encon Aquarion Eyewash Refill Cartridge
Dust cover for Fend-all Personal Eyewash Stations, single and double s (32-000517-0000).
More Details
Honeywell Accessories for Eyewashes
Buy Honeywell Accessories for Eyewashes Items Including:
Honeywell Personal Eyewash Wall Stations, 16 oz refill bottle.
More Details
Bradley Accessories for Eyewashes
Buy Bradley Accessories for Eyewashes Items Including:
Optional waste cart, 22-1/8"W x 33"H x 29-3/4"D.
More Details
Fend-all Accessories for Eyewashes
Buy Fend-all Accessories for Eyewashes Items Including:
Fend-all 2000™ Eyewash Refill Crtridge (32-ST2050-0000)
pH balanced, bacteria-inhibiting saline eyewash solution (32-000513-0000)
Bacteria-inhibiting eyewash preservative (32-001100-0000).
More Details
Fend all Eyewash 32 Oz
Buy Fend all Eyewash 32 Oz Items Including:
Fend-all Primary Eyewash Station (32-000200-0000)
Bacteria-inhibiting eyewash preservative (32-001100-0000)
Fend-all 2000™ Eyewash Refill Crtridge (32-ST2050-0000)
Universal cart, mount and transport most portable eyewash stations (32-001060-0000).
More Details
Product Type Eyewash Accessories
Buy Product Type Eyewash Accessories Items Including:
Encon Aquarion Eyewash Refill Cartridge
Honeywell Personal Eyewash Wall Stations, 16 oz refill bottle
Bacteria-inhibiting eyewash preservative (32-001100-0000)
Dust cover for Fend-all Personal Eyewash Stations, single and double s (32-000517-0000).
More Details
Eyewash Solutions
Buy Eyewash Solutions Items Including:
pH balanced, bacteria-inhibiting saline eyewash solution (32-000513-0000).
More Details
16 Refill
Buy 16 Refill Items Including:
Pipette Tips, Finntip-Flex, 100-1000uL, Refill Kit, (16) 96/pack refil (94060717).
More Details
Portable Cart
Buy Portable Cart Items Including:
Optional Four Wheel Cart
Universal cart, mount and transport most portable eyewash stations (32-001060-0000).
More Details
Fend Eyewash
Buy Fend Eyewash Items Including:
Bacteria-inhibiting eyewash preservative (32-001100-0000)
Fend-all 2000™ Eyewash Refill Crtridge (32-ST2050-0000)
Fend-all Primary Eyewash Station (32-000200-0000)
Universal cart, mount and transport most portable eyewash stations (32-001060-0000).
More Details
Eyewash Refills
Buy Eyewash Refills Items Including:
Encon Aquarion Eyewash Refill Cartridge
Scienceware Plum Sterile Saline Eyewash Refill, 200 mL, 3 bottles/pack
Scienceware Plum Sterile Saline Eyewash Refill, 500 mL, 3 bottles/pack
Honeywell Personal Eyewash Wall Stations, 16 oz refill bottle.
More Details
Fend Safety Products
Buy Fend Safety Products Items Including:
Fend-all Primary Eyewash Station (32-000200-0000)
pH balanced, bacteria-inhibiting saline eyewash solution (32-000513-0000)
Bacteria-inhibiting eyewash preservative (32-001100-0000)
Pureflow refill cartridges, premixed
Set of two cartridges..
More Details
100 Ml Freeze Dryer Adapter
Buy 100 Ml Freeze Dryer Adapter Items Including:
Fast-Freeze flask, 120 mL
Fast-Freeze flask, 150 mL
Fast-Freeze flask, 2000 mL
Adapter for 25, 50, and 100 mL Hamilton Gastight Syringe for use with (788314) (YO-74903-84)
Adapter for 25, 50, and 100 mL Hamilton Gastight Syringe for use with.
More Details
Accessories for Freezone Freeze-Dry Systems
Buy Accessories for Freezone Freeze-Dry Systems Items Including:
Fast-Freeze flask, 120 mL
Fast-Freeze flask, 150 mL
Fast-Freeze flask, 2000 mL
Clear Drying Chambers (YO-03337-64)
8 Port Clear Drying Chambers (YO-03337-66)
More Details
Fast Freezone Freeze Dry Systems
Buy Fast Freezone Freeze Dry Systems Items Including:
Fast-Freeze flask, 40 mL
Fast-Freeze flask, 80 mL
Fast-Freeze flask, 120 mL
Fast-Freeze flask, 150 mL
Fast-Freeze flask, 300 mL
More Details
Fast-Freeze Accessories for Freezone Freeze-Dry Systems
Buy Fast-Freeze Accessories for Freezone Freeze-Dry Systems Items Including:
Fast-Freeze flask, 40 mL
Fast-Freeze flask, 80 mL
Fast-Freeze flask, 120 mL
Fast-Freeze flask, 150 mL
Fast-Freeze flask, 300 mL
More Details
Fast-Freeze Freezone Freeze Dry Systems
Buy Fast-Freeze Freezone Freeze Dry Systems Items Including:
Fast-Freeze flask, 40 mL
Fast-Freeze flask, 80 mL
Fast-Freeze flask, 120 mL
Fast-Freeze flask, 150 mL
Fast-Freeze flask, 300 mL
More Details
Fast-Freeze Flasks
Buy Fast-Freeze Flasks Items Including:
Fast-Freeze flask, 40 mL
Fast-Freeze flask, 80 mL
Fast-Freeze flask, 120 mL
Fast-Freeze flask, 150 mL
Fast-Freeze flask, 300 mL
More Details
Fast-Freeze Freeze Dryers
Buy Fast-Freeze Freeze Dryers Items Including:
Fast-Freeze flask, 40 mL
Fast-Freeze flask, 80 mL
Fast-Freeze flask, 120 mL
Fast-Freeze flask, 150 mL
Fast-Freeze flask, 300 mL
More Details
Fast-Freeze Lab Equipment
Buy Fast-Freeze Lab Equipment Items Including:
Fast-Freeze flask, 40 mL
Fast-Freeze flask, 80 mL
Fast-Freeze flask, 120 mL
Fast-Freeze flask, 150 mL
Fast-Freeze flask, 300 mL
More Details
70618
results
<<<Previous
16
17
18
19
20
21
22
23
24
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cole-Parmer Accessories for Dredges to Fast-Freeze Lab Equipment
>
31125329-s-toc_2 31125329stoc2