Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 2354

Black and White Plates   to   Cole Cell Biology

Image
  Product Detail
More Detail

Black and White Plates

Buy Black and White Plates Items Including:
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well White Solid Plates, High binding
  • Corning 96-Well Black Solid Plates, Nontreated
  • Corning 96-Well Black Solid Plates, High binding
  • Plate,treated,black,100/cs (YO-01728-62).

Corning 96-Well Solid Plates

Buy Corning 96-Well Solid Plates Items Including:
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well White Solid Plates, High binding
  • Costar 96-Well Microplates, Immunology, PS, Flat, High Binding
  • Plate, 96 Well,ps,flat,100/cs (YO-01728-01)
  • Plate,96 Well,v-bottom,100/cs (YO-01728-05).

Thermo Scientific Nunc Cell Biology

Buy Thermo Scientific Nunc Cell Biology Items Including:
  • Thermo Scientific Nunc 96-Well Microplates, Nontreated, PP, Conical pl (YO-01929-30)
  • Thermo Scientific Nunc™ 96-Well Microplates, Nontreated, PP, Round, (YO-01929-32)
  • Thermo Scientific Nunc™ 96-Well Microplates, Nontreated, PP, Round, (YO-01929-33).

Corning 96-Well Black and White

Buy Corning 96-Well Black and White Items Including:
  • Corning 96-Well Black Solid Plates, Nontreated
  • Corning 96-Well Black Solid Plates, High binding
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well White Solid Plates, High binding
  • Plate,treated,black,100/cs (YO-01728-62).

Corning 96 Well Plates Half Areacls3679

Buy Corning 96 Well Plates Half Areacls3679 Items Including:
  • Corning 96-Well White Solid Plates, High binding
  • Corning 96-Well Clear-Bottom Plates; Black, Cell Culture Surface
  • Corning 96-Well Clear-Bottom Plates; White, Cell Culture Surface
  • Costar 96-Well Microplates, Immunology, PS, Round, Medium Binding.

Deep Well Plates

Buy Deep Well Plates Items Including:
  • 96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-04)
  • 96-well Deep Well Plate, Polystyrene, 32/pack (YO-07904-06)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-08).

Thermo Scientific Nunc Immunochemistry

Buy Thermo Scientific Nunc Immunochemistry Items Including:
  • Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-22)
  • Thermo Scientific Nunc™ Immunoplates; surface, nontreated, nonsteri (YO-01928-02)
  • Thermo Scientific Nunc™ Immunoplates; surface, nontreated, sterile; (YO-01928-04).

Cole 96-Well Deep Well Plates

Buy Cole 96-Well Deep Well Plates Items Including:
  • 96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-04)
  • 96-well Deep Well Plate, Polystyrene, 32/pack (YO-07904-06).

Polypropylene Plates

Buy Polypropylene Plates Items Including:
  • 96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-04)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-08)
  • Costar 96-Well Standard Microplates, Nontreated, PP, V-Bottom.

48 Well Plate Spinner

Buy 48 Well Plate Spinner Items Including:
  • 96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
  • Costar 48-well Multiple-well cell culture plates with lid, treated, st.

Storage Plates

Buy Storage Plates Items Including:
  • Storage Mat for Costar 96-Well Polypropylene Storage System
  • Costar 96-Well Standard Microplates, Nontreated, PP, Flat Well
  • Costar 96-Well Standard Microplates, Nontreated, PP, Round Well
  • Costar 96-Well Assay Blocks, Nontreated, PP, 1.0 mL, Sterile.

Thermo Scientific Nunc 96-Well

Buy Thermo Scientific Nunc 96-Well Items Including:
  • Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-22)
  • Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-32)
  • Thermo Scientific Nunc 96-Well Microplates, Nontreated, PP, Conical pl (YO-01929-30).

Corning Cell Biology

Buy Corning Cell Biology Items Including:
  • Corning 96-Well White Solid Plates, Nontreated
  • Plate,round,white,100/cs (YO-01728-60)
  • Plate,round,black,100/cs (YO-01728-61)
  • Plate,treated,black,100/cs (YO-01728-62)
  • Plate,treated,white,100/cs (YO-01728-63)

Corning Life Science

Buy Corning Life Science Items Including:
  • Corning 96-Well White Solid Plates, High binding
  • Corning cell culture dish, TC-treated, 35 mm diameter, 500/cs
  • Corning cell culture dish, TC-treated, 100 mm diameter, 500/cs
  • Corning cell culture flask, 75 cm2, straight neck, phenolic (YO-01835-12).

48 Well Plate

Buy 48 Well Plate Items Including:
  • 96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
  • Costar 48-well Multiple-well cell culture plates with lid, treated, st.

Assay Systems

Buy Assay Systems Items Including:
  • Costar 96-Well Assay Blocks, Nontreated, PP, 1.0 mL, Sterile
  • Costar 96-Well Assay Blocks, Nontreated, PP, 2.0 mL, Sterile
  • Costar 96-Well Assay Blocks, Nontreated, PP, 1.0 mL, Nonsterile.

Thermo Scientific Nunc Immunology

Buy Thermo Scientific Nunc Immunology Items Including:
  • Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-22)
  • Thermo Scientific Nunc™ Immunoplates; surface, nontreated, nonsteri (YO-01928-02)
  • Thermo Scientific Nunc™ Immunoplates; surface, nontreated, sterile; (YO-01928-04).

Cell Biology Products

Buy Cell Biology Products Items Including:
  • Plate,treated,black,100/cs (YO-01728-62)
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well White Solid Plates, High binding
  • Corning 96-Well Black Solid Plates, Nontreated
  • Costar 96-Well Standard Microplates, Nontreated, PP, Flat Well.

Cell Biology Products Manufacturers

Buy Cell Biology Products Manufacturers Items Including:
  • Plate,treated,black,100/cs (YO-01728-62)
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well White Solid Plates, High binding
  • Corning 96-Well Black Solid Plates, Nontreated
  • Costar 96-Well Standard Microplates, Nontreated, PP, Flat Well.

Corning 96-Well Solid P

Buy Corning 96-Well Solid P Items Including:
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well Black Solid Plates, Nontreated
  • Costar 96-Well Microplates, Immunology, PS, Flat, Medium Binding
  • Plate, 96 Well,ps,flat,100/cs (YO-01728-01)
  • Plate,96 Well,v-bottom,100/cs (YO-01728-05).

Cell Biology

Buy Cell Biology Items Including:
  • Thermo Scientific Nunc™ 96-Well Microplates, Nontreated, PP, Round, (YO-01929-32)
  • Thermo Scientific Nunc™ 96-Well Microplates, Nontreated, PP, Round, (YO-01929-33)
  • Thermo Scientific Nunc 96-Well Microplates, Nontreated, PP, Conical pl (YO-01929-34).

Well Shape Conical

Buy Well Shape Conical Items Including:
  • Thermo Scientific Nunc 96-Well Microplates, Nontreated, PP, Conical pl (YO-01929-38)
  • Thermo Scientific Nunc 96-Well Microplates, Nontreated, PP, Conical pl (YO-01929-30).

Cole-Parmer Cell Biology

Buy Cole-Parmer Cell Biology Items Including:
  • Storage Mat Applicator (YO-01729-24)
  • 96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-04)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-08)
  • E-Vac Aspiration System.

Well Volume(Ml) 0.3

Buy Well Volume(Ml) 0.3 Items Including:
  • Thermo Scientific Nunc™ Immunoplates; surface, Maxisorp; well shape
  • Thermo Scientific Nunc 96-Well Microplates, Nontreated, PP, Conical pl (YO-01929-30)
  • Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 96-well Micro (YO-01930-10).

Biology Life Science

Buy Biology Life Science Items Including:
  • Plate,treated,black,100/cs (YO-01728-62)
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well White Solid Plates, High binding
  • Corning 96-Well Black Solid Plates, Nontreated
  • Costar 96-Well Standard Microplates, Nontreated, PP, Flat Well.

Nunc Immunology

Buy Nunc Immunology Items Including:
  • Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-22)
  • Thermo Scientific Nunc™ Immunoplates; surface, nontreated, nonsteri (YO-01928-02)
  • Thermo Scientific Nunc™ Immunoplates; surface, nontreated, sterile; (YO-01928-04).

Biology Cell Biology

Buy Biology Cell Biology Items Including:
  • Plate,treated,black,100/cs (YO-01728-62)
  • Corning 96-Well White Solid Plates, Nontreated
  • Corning 96-Well White Solid Plates, High binding
  • Corning 96-Well Black Solid Plates, Nontreated
  • Costar 96-Well Standard Microplates, Nontreated, PP, Flat Well.

Cole Cell Biology

Buy Cole Cell Biology Items Including:
  • Storage Mat Applicator (YO-01729-24)
  • 96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-04)
  • 96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-08)
  • E-Vac Aspiration System.

70618 results


Black and White Plates to Cole Cell Biology

>