Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 25
Pomona Modular Passive Oscilloscope Probes
to
Electrical Standards
Image
Product Detail
More Detail
Pomona Modular Passive Oscilloscope Probes
Buy Pomona Modular Passive Oscilloscope Probes Items Including:
6498: Modular Passive Oscilloscope Probe, 100:1 Ratio
6499: Modular Passive Oscilloscope Probe, 1:1 Ratio to 10:1
6500: Modular Passive Oscilloscope Probe, 1:1 Ratio to 10:1
6553: Modular Oscilloscope Probe Tip Adapter to SMD Grabber Adapter.
More Details
Davis Tektronix Oscilloscope Accessories & Probes
Buy Davis Tektronix Oscilloscope Accessories & Probes Items Including:
GPIB To USB Adapter for Tektronix Oscilloscopes (TEK-USB-488)
Hard Transit Carrying Case (requires 20013-44) for Tektronix Oscillosc (HCTEK4321)
Large Nylon Soft Carrying Case for Tektronix Series Oscilloscopes.
More Details
Tektronix Accessories for Oscilloscopes
Buy Tektronix Accessories for Oscilloscopes Items Including:
Differential Preamplifier for TDS3000B Series Oscilloscopes (requires (ADA400A)
Standard Passive Probe (10X, 100 MHz) for TDS3000B Series Oscilloscope (P3010)
Tektronix Current Probe, 120 MHz, 100A, 5 FT w/ Term Safety Controlled (P6022).
More Details
Davis Accessories for Oscilloscopes
Buy Davis Accessories for Oscilloscopes Items Including:
Limit Testing Module for Tektronix TDS3000B Series Oscilloscopes (YO-20013-57)
Telcom Mask Testing Module for Tektronix TDS3000B Series Oscilloscopes (YO-20013-60)
Plug-In Communication Module for Tektronix TDS3000B Series Oscilloscop (YO-20013-71).
More Details
Tektronix Oscilloscopes Probes
Buy Tektronix Oscilloscopes Probes Items Including:
Tektronix Current Probe, 120 MHz, 100A, 5 FT w/ Term Safety Controlled (P6022)
Tektronix Current Probe, 60 MHz, 250A 5FT w/ Term Safety Controlled (P6021)
Standard Passive Probe (10X, 100 MHz) for TDS3000B Series Oscilloscope (P3010).
More Details
Cole-Parmer Oscium Imso-104 Oscilloscope Accessories
Buy Cole-Parmer Oscium Imso-104 Oscilloscope Accessories Items Including:
Oscium OSC1100 Analog Probe, 1x / 10x
Oscium OSC3055 Logic Harness, 5 Pin with SMD Grabbers
Oscium OSC3050 Logic Harness, 5 Pin
Oscium OSC3005 SMD Grabbers
Oscium OSC1109 Extension cable, 19 in..
More Details
Tektronix Current Probes
Buy Tektronix Current Probes Items Including:
Tektronix Current Probe, 120 MHz, 100A, 5 FT w/ Term Safety Controlled (P6022)
Tektronix Current Probe, 60 MHz, 250A 5FT w/ Term Safety Controlled (P6021)
Tektronix AC/DC Current Probe 100A, 100 KHz
TCP0150, TekVPI DC to 20MHz, 150 A Current Probe.
More Details
Pomona Accessories for Oscilloscopes
Buy Pomona Accessories for Oscilloscopes Items Including:
6493: Pomona Scope Probes
6498: Modular Passive Oscilloscope Probe, 100:1 Ratio
6495: Modular Passive Oscilloscope Probe, 10:1 Ratio
6496: Modular Passive Oscilloscope Probe, 10:1 Ratio
6499: Modular Passive Oscilloscope Probe, 1:1 Ratio to 10:1.
More Details
Cole-Parmer Oscilloscopes
Buy Cole-Parmer Oscilloscopes Items Including:
Oscium OSC1100 Analog Probe, 1x / 10x
Oscium OSC3055 Logic Harness, 5 Pin with SMD Grabbers
Oscium OSC3050 Logic Harness, 5 Pin
Oscium OSC3005 SMD Grabbers
Oscium OSC1109 Extension cable, 19 in..
More Details
Tektronix Probes
Buy Tektronix Probes Items Including:
Standard Passive Probe (10X, 100 MHz) for TDS3000B Series Oscilloscope (P3010)
High Voltage Differential Probe (50X/500X, 100 MHz) for TDS3000B Serie (P5205)
Standard Passive Probe with Readout (10X, 500 MHz) for TDS3000B Series (P6139A).
More Details
Oscium Oscilloscopes
Buy Oscium Oscilloscopes Items Including:
Oscium Logic Harness, OSC3056, 9 Pin with SMD Grabbers
Oscium Conducted Measurement Kit, OSC1101 for WiPry-Combo and WiPry-Po
Oscium iMSO-104 Oscilloscope for iPad, iPhone, iPod, 5 MHz, 12 MS/s
Oscium OSC3055 Logic Harness, 5 Pin with SMD Grabbers.
More Details
Pomona Oscilloscopes
Buy Pomona Oscilloscopes Items Including:
6493: Pomona Scope Probes
6498: Modular Passive Oscilloscope Probe, 100:1 Ratio
6495: Modular Passive Oscilloscope Probe, 10:1 Ratio
6496: Modular Passive Oscilloscope Probe, 10:1 Ratio
6499: Modular Passive Oscilloscope Probe, 1:1 Ratio to 10:1.
More Details
Probe High Voltage
Buy Probe High Voltage Items Including:
Bartol Mag-Probe High Sensitivity Noncontact Probe
Tektronix High Voltage Single-ended Probe, 100X, 250 MHz (P5100)
Tektronix High Voltage Single-ended Probe, 1000X, 75 MHz, 10 FT (P6015A)
Tektronix TPP0850 High Voltage Probe; 50X, 800 MHZ, 2.5 kV.
More Details
Oscilloscope Accessories
Buy Oscilloscope Accessories Items Including:
Active Probe (10X, 1 GHz) for TDS3000B Series Oscilloscopes (P6243)
Large Nylon Soft Carrying Case for Tektronix Series Oscilloscopes
6498: Modular Passive Oscilloscope Probe, 100:1 Ratio
6499: Modular Passive Oscilloscope Probe, 1:1 Ratio to 10:1.
More Details
Active Probe
Buy Active Probe Items Including:
TekVPI 500 MHz differential active probe (TDP0500)
Handheld Meter Kit, 14-mm sample cup size, one probe input (one probe (HYGROPALM-AW1 W/14ML CUP)
Handheld Meter Kit, 40-mm sample cup size, one probe input (one probe (HYGROPALM-AW1 W/40ML CUP).
More Details
Tektronix Dc
Buy Tektronix Dc Items Including:
TCP404XL : Probe AC/DC Current DC-2MHz 500A DC (TCP404XL)
TCPA300 : Amplifier AC/DC Current Probe DC-100MHz (TCPA300)
Tektronix PWS4205 Programmable DC Power Supply, 18 V, 5 A
Tektronix PWS4602 Programmable DC Power Supply, 60 V, 2.5 A.
More Details
Probe Amplifier
Buy Probe Amplifier Items Including:
TCPA300 : Amplifier AC/DC Current Probe DC-100MHz (TCPA300).
More Details
Product Type Oscilloscope Probe
Buy Product Type Oscilloscope Probe Items Including:
6498: Modular Passive Oscilloscope Probe, 100:1 Ratio
Large Nylon Soft Carrying Case for Tektronix Series Oscilloscopes
Tektronix Probe Amplifier AC/DC Current, 50 MHz (TCPA400)
Tektronix Passive Probe, Volt 1X/10X, 200 MHz, 300V 30V Float (P2220).
More Details
Pomona Modular Passive Oscilloscope
Buy Pomona Modular Passive Oscilloscope Items Including:
6498: Modular Passive Oscilloscope Probe, 100:1 Ratio
6495: Modular Passive Oscilloscope Probe, 10:1 Ratio
6496: Modular Passive Oscilloscope Probe, 10:1 Ratio
6499: Modular Passive Oscilloscope Probe, 1:1 Ratio to 10:1
6493: Pomona Scope Probes.
More Details
Tektronix Current
Buy Tektronix Current Items Including:
Tektronix Current Probe, 60 MHz, 250A 5FT w/ Term Safety Controlled (P6021)
TCP404XL : Probe AC/DC Current DC-2MHz 500A DC (TCP404XL)
TCP305 : Probe AC/DC Current DC-50MHz 50A DC (TCP305)
TCPA300 : Amplifier AC/DC Current Probe DC-100MHz (TCPA300).
More Details
Davis Oscilloscopes
Buy Davis Oscilloscopes Items Including:
Limit Testing Module for Tektronix TDS3000B Series Oscilloscopes (YO-20013-57)
Telcom Mask Testing Module for Tektronix TDS3000B Series Oscilloscopes (YO-20013-60)
Plug-In Communication Module for Tektronix TDS3000B Series Oscilloscop (YO-20013-71).
More Details
Oscilloscope Voltage
Buy Oscilloscope Voltage Items Including:
High Voltage Differential Probe (50X/500X, 100 MHz) for TDS3000B Serie (P5205)
Tektronix High Voltage Differential Probe, 50X/500X, 25 MHz, Power 1K (P5200)
Fluek VPS410-B, Voltage probe set 10:1, blue
Fluek VPS410-V, Voltage probe set 10:1, green.
More Details
B & K Oscilloscopes
Buy B & K Oscilloscopes Items Including:
Active Probe (10X, 1 GHz) for TDS3000B Series Oscilloscopes (P6243)
P6245 : Active FET Probe 1.5GHz 10X 1P TDS Series Oscilloscopes (P6245)
6498: Modular Passive Oscilloscope Probe, 100:1 Ratio
6495: Modular Passive Oscilloscope Probe, 10:1 Ratio.
More Details
Tektronix 50
Buy Tektronix 50 Items Including:
High Voltage Differential Probe (100X/1000X, 50 MHz) for TDS3000B Seri
Tektronix AC Current Probe 1000 ARMS, 50 KHz
Tektronix AFG3051C Arbitrary Function Generator, 1 Channel, 50 MHz
Tektronix PA4000 Multi-phase Power Analyzer, One Input Channel.
More Details
100 Oscilloscope
Buy 100 Oscilloscope Items Including:
Standard Passive Probe (10X, 100 MHz) for TDS3000B Series Oscilloscope (P3010)
Tektronix TDS2012B, Color Digital Storage Oscilloscope, 2 Channel, 100 (YO-20013-87)
Tektronix TDS2014B, Color Digital Storage Oscilloscope, 4 Channel, 100 (YO-20013-88).
More Details
Oscium Process Equipment & Instrumentation
Buy Oscium Process Equipment & Instrumentation Items Including:
Oscium Conducted Measurement Kit, OSC1101 for WiPry-Combo and WiPry-Po
Oscium iMSO-104 Oscilloscope for iPad, iPhone, iPod, 5 MHz, 12 MS/s
Oscium WiPry-Power, OSC1002, Dynamic Power Meter for iPhone, iPad, and
Oscium OSC1100 Analog Probe, 1x / 10x.
More Details
Davis Process Equipment & Instrumentation
Buy Davis Process Equipment & Instrumentation Items Including:
Hard Transit Carrying Case (requires 20013-44) for Tektronix Oscillosc (HCTEK4321)
GPIB To USB Adapter for Tektronix Oscilloscopes (TEK-USB-488)
TekVPI To TekProbe BNC Adapter (TPA-BNC)
Tektronix Probe Amplifier AC/DC Current, 50 MHz (TCPA400).
More Details
Tek Tektronix
Buy Tek Tektronix Items Including:
Deskew Pulse Generator Signal Source w/TekVPITM Scope Interface (TEK-DPG).
More Details
Tektronix 100
Buy Tektronix 100 Items Including:
Standard Passive Probe (10X, 100 MHz) for TDS3000B Series Oscilloscope (P3010)
Tektronix TDS2012B, Color Digital Storage Oscilloscope, 2 Channel, 100 (YO-20013-87)
Tektronix TDS2014B, Color Digital Storage Oscilloscope, 4 Channel, 100 (YO-20013-88).
More Details
Electrical Standards
Buy Electrical Standards Items Including:
Standard Passive Probe (10X, 100 MHz) for TDS3000B Series Oscilloscope (P3010)
Standard Passive Probe with Readout (10X, 500 MHz) for TDS3000B Series (P6139A)
Standard 6-outlet strip, 15-ft cord (UL207BD)
Standard Analog/Digital Megohmmeter (1040).
More Details
70618
results
<<<Previous
21
22
23
24
25
26
27
28
29
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Pomona Modular Passive Oscilloscope Probes to Electrical Standards
>
31125329-s-toc_2 31125329stoc2