Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 26
Oscium Electrical
to
1.5
Image
Product Detail
More Detail
Oscium Electrical
Buy Oscium Electrical Items Including:
Oscium Conducted Measurement Kit, OSC1101 for WiPry-Combo and WiPry-Po
Oscium iMSO-104 Oscilloscope for iPad, iPhone, iPod, 5 MHz, 12 MS/s
Oscium WiPry-Power, OSC1002, Dynamic Power Meter for iPhone, iPad, and
Oscium OSC1100 Analog Probe, 1x / 10x.
More Details
Davis Electrical
Buy Davis Electrical Items Including:
Hard Transit Carrying Case (requires 20013-44) for Tektronix Oscillosc (HCTEK4321)
GPIB To USB Adapter for Tektronix Oscilloscopes (TEK-USB-488)
TekVPI To TekProbe BNC Adapter (TPA-BNC)
Tektronix Probe Amplifier AC/DC Current, 50 MHz (TCPA400).
More Details
Benchtop Stand
Buy Benchtop Stand Items Including:
Benchtop Stand for PressureMAT Monitor
Oakton CON 700 Meter w/probe & stand
Oakton pH 700 Benchtop Meter, meter and probe stand
Oakton PC2700 Meter and electrode stand
Oakton CON 2700 meter w/probe & stand with NIST-traceable calibvration.
More Details
Accessories for Panel-Mount Meters
Buy Accessories for Panel-Mount Meters Items Including:
Benchtop stand for 1/8 DIN panel meters (05656-55/CLR-00001DE FEET)
MBLPAX00:Lpax Mounting Bracket.
More Details
Panel Meters Accessories
Buy Panel Meters Accessories Items Including:
Benchtop stand for 1/8 DIN panel meters (05656-55/CLR-00001DE FEET)
Heavy-Duty Wall Mount Bracket (for panel meter only) (3-0000.596)
Meter Copy Cable
4 Relays Expansion Module
4 Digital Inputs & 4 Outputs
More Details
Cole-Parmer Moisture Remover
Buy Cole-Parmer Moisture Remover Items Including:
Moisture remover, 220 VAC (800-DT/EXP).
More Details
Accessories for Plas-Labs Glove Boxes
Buy Accessories for Plas-Labs Glove Boxes Items Including:
Iris diaphragm retrofit kit; order to fit your current glove box (800/IRIS/CONV)
Iris diaphragm, 6" diameter (800-IRIS)
Moisture remover, 115 VAC (800-DT)
Moisture remover, 220 VAC (800-DT/EXP)
Catalyst heater; 115 VAC (800-HEATER).
More Details
Heater Boxes
Buy Heater Boxes Items Including:
Catalyst heater, 220 VAC (800-HEATER/EXP).
More Details
Moisture Remover
Buy Moisture Remover Items Including:
Moisture remover, 220 VAC (800-DT/EXP).
More Details
Cole Moisture Remover
Buy Cole Moisture Remover Items Including:
Moisture remover, 220 VAC (800-DT/EXP).
More Details
Iris Diaphragm
Buy Iris Diaphragm Items Including:
Iris diaphragm retrofit kit; order to fit your current glove box (800/IRIS/CONV).
More Details
Cole-Parmer Accessories for Plas-Labs Glove Boxes
Buy Cole-Parmer Accessories for Plas-Labs Glove Boxes Items Including:
Plas Labs Glove Box Pressure Relief Valve, 1/8" NPT Stainless Steel
Moisture remover, 115 VAC (800-DT)
Moisture remover, 220 VAC (800-DT/EXP)
Catalyst heater; 115 VAC (800-HEATER).
More Details
Cole-Parmer Catalyst Heater
Buy Cole-Parmer Catalyst Heater Items Including:
Catalyst heater, 220 VAC (800-HEATER/EXP).
More Details
Catalyst Heater
Buy Catalyst Heater Items Including:
Catalyst heater, 220 VAC (800-HEATER/EXP).
More Details
Cole Catalyst Heater
Buy Cole Catalyst Heater Items Including:
Catalyst heater, 220 VAC (800-HEATER/EXP).
More Details
Peristaltic Pump Pulse Dampener
Buy Peristaltic Pump Pulse Dampener Items Including:
PTFE Pulse Dampeners With PTFE Diaphragm, 3/8" NPT
PTFE Pulse Dampeners With PTFE Diaphragm, 3/4" NPT
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 1-1/2" NPT
High-Flow Pulse Dampener for Series Q piston pump heads (PDHF).
More Details
Small Continuous Liquid Diaphragm Pump
Buy Small Continuous Liquid Diaphragm Pump Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Pump, Metering with 5 control methods, 0.66gph, PVDF, Fkm Front Contro (96470133)
Pump, Metering with 5 control methods, 4.88gph, PVDF, Fkm Front Contro (96470352).
More Details
500 Psi Water Pump
Buy 500 Psi Water Pump Items Including:
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Integrated Pressure Coolant System, 8 GPM, 500 PSI, Brass head constru
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade.
More Details
Santoprene Diaphragm
Buy Santoprene Diaphragm Items Including:
Air-Powered Double-Diaphragm Pump, 1/4" Connections, PP / Santoprene w
High-Flow Double-Diaphragm Pump, 1/2" Connections, PP / Santoprene wet
Air-Powered Double-Diaphragm Pump, 1/4" Connections, Acetal / Santopre.
More Details
Liquid Pump Types
Buy Liquid Pump Types Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
High-Purity Liquid Centrifugal Pump, 316L Stainless Steel, 1 Hp, 10 GP.
More Details
Water Level Controller Circuit
Buy Water Level Controller Circuit Items Including:
WLT100 : Water Level Indicator 100FT
(WLT100)
WLT200 : Water Level Indicator 200FT (WLT200)
WLT300 : Water Level Indicator 300FT (WLT300)
WLT500 : Water Level Indicator 500FT (WLT500)
More Details
Lab Pump Junior Model Rhsy
Buy Lab Pump Junior Model Rhsy Items Including:
Service kit for models 75300-02
Service kit for models 75300-12
Service kit for models 75300-04
Service kit for models 75300-14
Service kit for model 75320-60
More Details
Stainless Steel Pressure Relief Valve
Buy Stainless Steel Pressure Relief Valve Items Including:
Plas Labs Glove Box Pressure Relief Valve, 1/8" NPT Stainless Steel
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Unloader (Pressure Relief) Valve, Stainless Steel, 500 to 2500 PSI Sta.
More Details
Miniature Liquid Diaphragm Pump
Buy Miniature Liquid Diaphragm Pump Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 1.5 LPM, 12 VDC, GAS ONLY,.
More Details
Pumping Valve
Buy Pumping Valve Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Unloader (Pressure Relief) Valve, Stainless Steel, 500 to 2500 PSI Sta.
More Details
Coolant Pump Output Brass Fitting
Buy Coolant Pump Output Brass Fitting Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Integrated Pressure Coolant System, 8 GPM, 500 PSI, Brass head constru
Integrated Pressure Coolant System, 8 GPM, 1000 PSI, Brass head constr.
More Details
Stainless Steel Pumps and Valves
Buy Stainless Steel Pumps and Valves Items Including:
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Unloader (Pressure Relief) Valve, Stainless Steel, 500 to 2500 PSI Sta
316 stainless steel 90° full-cone spray nozzle, 0.053" orifice.
More Details
Design Valve Micro Diaphragm Pump
Buy Design Valve Micro Diaphragm Pump Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Cole-Parmer Inert Solenoid Self Priming Micro Pumps, 20 microL, 12 VDC (120SP1220-4EE)
Cole-Parmer Inert Solenoid Self Priming Micro Pumps, 20 microL, 24 VDC (120SP2420-4EE).
More Details
Liquid Operated Ventura Liquid Pump
Buy Liquid Operated Ventura Liquid Pump Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC.
More Details
1.5 Inch Liquid Level Controller
Buy 1.5 Inch Liquid Level Controller Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
Flowline Switch-Pro Remote Isolation Relay Level Controller, 2 SPDT re.
More Details
70618
results
<<<Previous
22
23
24
25
26
27
28
29
30
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Oscium Electrical to 1.5
>
31125329-s-toc_2 31125329stoc2