Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 28
Pumps Psi
to
Qsonica Lab Equipment
Image
Product Detail
More Detail
Pumps Psi
Buy Pumps Psi Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Unloader (Pressure Relief) Valve, Stainless Steel, 500 to 2500 PSI Sta.
More Details
Pump Hi Pressure
Buy Pump Hi Pressure Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Unloader (Pressure Relief) Valve, Stainless Steel, 500 to 2500 PSI Sta.
More Details
Lmi Liquid Level Controller
Buy Lmi Liquid Level Controller Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
ExOsense Liquid Level Sensor Assembly; High-Level, Wet Sink, PVC (219003).
More Details
Kimray Liquid Level Controller
Buy Kimray Liquid Level Controller Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
ExOsense Liquid Level Sensor Assembly; High-Level, Wet Sink, PVC (219003).
More Details
High Speed High Pressure Pumps
Buy High Speed High Pressure Pumps Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Unloader (Pressure Relief) Valve, Stainless Steel, 500 to 2500 PSI Sta.
More Details
High Volume High Pressure Pumps
Buy High Volume High Pressure Pumps Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Unloader (Pressure Relief) Valve, Stainless Steel, 500 to 2500 PSI Sta.
More Details
Liquid Level Controllers for Air-Operate
Buy Liquid Level Controllers for Air-Operate Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC.
More Details
Cole Liquid Level Controllers for Air-Operate
Buy Cole Liquid Level Controllers for Air-Operate Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC.
More Details
Level Liquid
Buy Level Liquid Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
WLT100 : Water Level Indicator 100FT
(WLT100)
WLT200 : Water Level Indicator 200FT (WLT200)
ExOsense Liquid Level Sensor Assembly; High-Level, Wet Sink, PVC (219003).
More Details
Stainless Pumps
Buy Stainless Pumps Items Including:
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST562P13F)
Stainless Steel NEMA Type C-face Rigid Base Single-phase Motor, 1/3 Hp (1ST564P13F).
More Details
Cole Accessories for Positive Displacement Pumps
Buy Cole Accessories for Positive Displacement Pumps Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0).
More Details
Product Type Service Kits
Buy Product Type Service Kits Items Including:
Service kit for models 75300-00, -20
Service kit for models 75300-02
Service kit for models 75300-12
Service kit for models 75300-04
Service kit, includes electrolyte solution, pp membranes, O-rings, and
More Details
Positive Displacement Valve
Buy Positive Displacement Valve Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice.
More Details
Steel Supplies
Buy Steel Supplies Items Including:
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain
Unloader (Pressure Relief) Valve, Stainless Steel, 500 to 2500 PSI Sta
White peg for modular stainless steel drying racks, 12"L
Support Stand Stainless steel with 28” Rod.
More Details
Liquid to Liquid
Buy Liquid to Liquid Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Disposable In-Line Gas/Liquid Filters; Nylon; gas efficiency, 99.99%; (YO-02908-62)
Disposable In-Line Gas/Liquid Filters; Nylon; gas efficiency, 99.99%; (YO-02908-64).
More Details
Unloader Valves
Buy Unloader Valves Items Including:
Unloader (Pressure Relief) Valve, Brass, 75 to 500 PSI Brass Unloader
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain.
More Details
Cole-Parmer Level Control
Buy Cole-Parmer Level Control Items Including:
Cole-Parmer batch controller
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
Cole-Parmer Point Level Switch Pack; Two Switches, PP.
More Details
Cole Liquid Level Controller
Buy Cole Liquid Level Controller Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Single-point controller; 24 to 260 VAC, 5 A load (20173)
Economical liquid alarm (3240CP)
Electro-optic point-level sensor; nylon, wet sink, 1/4" NPT(M) (157750).
More Details
Pump Ad
Buy Pump Ad Items Including:
Hand-operated acid-transfer pump, 6 strokes/gallon, female buttress ad (236PF3P)
PTFE Pulse Dampeners With PTFE Diaphragm, 3/8" NPT
PTFE Pulse Dampeners With PTFE Diaphragm, 3/4" NPT
PTFE Pulse Dampeners With PTFE Diaphragm, 1" NPT.
More Details
Valves Supply
Buy Valves Supply Items Including:
Unloader (Pressure Relief) Valve, Brass, 500 to 2500 PSI Brass Unloade
Black polypropylene valve cap (154134)
Advantec Vacuum Filtration Manifold; PVC, 3-Branch with 2-Way Valves
Advantec Vacuum Filtration Manifold; PVC, 6-Branch with 2-Way Valves.
More Details
Cole Level Control
Buy Cole Level Control Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Level Sensor 1/4"npt-wet 12v
Level Sensor 1/4"npt-dry 12v
Level Sensor 1/2"-20-wet 12v
Level Sensor 1/2"-20-dry 12v
More Details
Pvc Air
Buy Pvc Air Items Including:
In-Line Direct Reading Flowmeter for Air; 20 to 240 scfm, PVC Fittings (FR5A81PI-D).
More Details
Ultrasonic Probe Sonicators
Buy Ultrasonic Probe Sonicators Items Including:
Microprobe, 5/64", 0.2 - 5ml, for use with Q55 Sonicator
Ultrasonic Cell Disruptor Replacement Tips, 1/2" dia
Ultrasonic Cell Disruptor Replacement Tips, 3/4" dia
Ultrasonic Cell Disruptor Replacement Tips, 1" dia.
More Details
Sonication Probes
Buy Sonication Probes Items Including:
Probe, 1", solid
Microprobe, 5/64", 0.2 - 5ml, for use with Q55 Sonicator
Microprobe, 1/8", 0.5 - 15ml, for use with Q55 Sonicator
Coupler (for Probe 04712-65) for use with ultrasonic processors
Probe, 1", with replaceable tip.
More Details
Qsonica Sonicator Microtip Probes
Buy Qsonica Sonicator Microtip Probes Items Including:
Ultrasonic Cell Disruptor Microtip Probes, 1-16" dia
Ultrasonic Cell Distruptor Microtip Probe 1/8" dia
Ultrasonic Cell Disruptor Microtip Probes, 1/4" dia.
More Details
Sonicators for Lab
Buy Sonicators for Lab Items Including:
Microprobe, 5/64", 0.2 - 5ml, for use with Q55 Sonicator
Microprobe, 1/8", 0.5 - 15ml, for use with Q55 Sonicator
Qsonica 55 Watt Sonicator, 110V
Qsonica 55 Watt Sonicator, 220V
Qsonica Touch-Screen Sonicator; 700 watt, 120VAC, 50/60 Hz.
More Details
Ultrasonic Horns
Buy Ultrasonic Horns Items Including:
Qsonica Ultrasonic Cell Disruptor Cup Horn Stand
Cup horn; 2-1/2" dia (YO-04710-38)
Micro Cup Horn (630-0447)
Cup horn (5.5") for use with ultrasonic processors
Dual Horn (with two 3/4" solid probes) for use with ultrasonic process.
More Details
Lab Sonication Probe
Buy Lab Sonication Probe Items Including:
Probe, 1/2", with replaceable tip
Probe, 1", with replaceable tip
Ultrasonic Cell Disruptor Microtip Probes, 1-16" dia
Ultrasonic Cell Distruptor Microtip Probe 1/8" dia
Ultrasonic Cell Disruptor Microtip Probes, 1/4" dia.
More Details
Sonic Accessorie
Buy Sonic Accessorie Items Including:
Microprobe, 5/64", 0.2 - 5ml, for use with Q55 Sonicator
Microprobe, 1/8", 0.5 - 15ml, for use with Q55 Sonicator
Microprobe, 1/4", 10 - 50ml, for use with Q55 Sonicator
QSonica Sonicator Continuous Flow Cell
Converter cable; 10-ft.
More Details
Qsonica Lab Equipment
Buy Qsonica Lab Equipment Items Including:
QSonica Sonicator Continuous Flow Cell
Qsonica Touch-Screen Sonicator; 700 watt, 120VAC, 50/60 Hz
Qsonica Touch-Screen Sonicator; 700 watt, 220VAC, 50/60 Hz
Qsonica 55 Watt Sonicator, 110V
Ultrasonic Cell Disruptor Replacement Tips, 1/2" dia.
More Details
70618
results
<<<Previous
24
25
26
27
28
29
30
31
32
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Pumps Psi to Qsonica Lab Equipment
>
31125329-s-toc_2 31125329stoc2