Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 30
Cole Beaker Positioning Covers
to
Revco Freezer Accessories
Image
Product Detail
More Detail
Cole Beaker Positioning Covers
Buy Cole Beaker Positioning Covers Items Including:
Beaker positioning cover for Cole-Parmer 1/2 Gallon and Stainless Stee
Beaker positioning cover for Cole-Parmer 3/4 Gallon and Stainless Stee
Beaker positioning cover for Cole-Parmer 1 1/2 Gallon and Stainless St.
More Details
Solid Insert
Buy Solid Insert Items Including:
Solid tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon C
Solid stainless steel pan for Cole-Parmer 3/4 Gallon and Stainless Ste
Solid stainless steel pan for Cole-Parmer 1 1/2 Gallon and Stainless S.
More Details
Cole Accessories for Standard Ultrasonic Cleaners
Buy Cole Accessories for Standard Ultrasonic Cleaners Items Including:
Solid tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon C
Perforated tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gal
Mesh tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon Cl.
More Details
Cole Mesh Insert Trays
Buy Cole Mesh Insert Trays Items Including:
Mesh tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon Cl
Mesh tray for Cole-Parmer 3/4 Gallon and Stainless Steel 1 Gallon Clea.
More Details
Cole Perforated Insert Trays
Buy Cole Perforated Insert Trays Items Including:
Perforated tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gal
Perforated tray for Cole-Parmer 3/4 Gallon and Stainless Steel 1 Gallo
Perforated tray for Cole-Parmer 1 1/2 Gallon and Stainless Steel 1 1/2.
More Details
Cole Solid Insert Trays
Buy Cole Solid Insert Trays Items Including:
Solid tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon C
Solid stainless steel pan for Cole-Parmer 3/4 Gallon and Stainless Ste
Solid stainless steel pan for Cole-Parmer 1 1/2 Gallon and Stainless S.
More Details
Cole-Parmer Standard Beakers
Buy Cole-Parmer Standard Beakers Items Including:
Beaker positioning cover for Cole-Parmer 1/2 Gallon and Stainless Stee
Beaker positioning cover for Cole-Parmer 3/4 Gallon and Stainless Stee
Beaker positioning cover for Cole-Parmer 1 1/2 Gallon and Stainless St.
More Details
Ultrasonic Insert
Buy Ultrasonic Insert Items Including:
Solid tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon C
Perforated tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gal
Mesh tray for Cole-Parmer 1/2 Gallon and Stainless Steel 3/4 Gallon Cl.
More Details
Stainless Perforated
Buy Stainless Perforated Items Including:
Sanitary In-Line Strainer Assembly; 1" Tri-Clamp; with 1/8" Perforated
Sanitary In-Line Strainer Assembly; 2" Tri-Clamp; with 1/8" Perforated
Sanitary In-Line Strainer Assembly; 1" Tri-Clamp; with 1/4" Perforated.
More Details
Cole-Parmer Pan
Buy Cole-Parmer Pan Items Including:
Solid stainless steel pan for Cole-Parmer 3/4 Gallon and Stainless Ste
Receiving pan for 200-mm sieves
Receiving Pan with Fitted Rim for Nesting 8" Full Height Brass Sieves
Receiving Pan for 8" Diameter Stainless Steel Full Height Sieves.
More Details
Cole Stainless Stee
Buy Cole Stainless Stee Items Including:
Cole-Parmer Hi-Accuracy RTD Freezer Thermometer with 1 Stainless Stee
Beaker positioning cover for Cole-Parmer 1/2 Gallon and Stainless Stee.
More Details
Thermo Scientific Revco Elite Plus Freezer
Buy Thermo Scientific Revco Elite Plus Freezer Items Including:
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-02)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03).
More Details
Revco Freezer
Buy Revco Freezer Items Including:
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-02)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03).
More Details
Thermo Revco Freezers
Buy Thermo Revco Freezers Items Including:
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 17.3 c (YO-44146-25)
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-26)
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-27).
More Details
Thermo Freezer
Buy Thermo Freezer Items Including:
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 17.3 c (YO-44146-25)
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-26)
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-27).
More Details
Thermo Scientific Revco Ultra-Low Temperature Freezers
Buy Thermo Scientific Revco Ultra-Low Temperature Freezers Items Including:
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-02)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03).
More Details
Revco Scientific
Buy Revco Scientific Items Including:
Thermo Scientific Revco Enzyme freezer, -20°C, 208/230 VAC, 60 Hz
Thermo Scientific Revco Lab freezer, -20°C, 115 VAC, 60 Hz
Thermo Scientific Revco Lab freezer, -20°C, 220 VAC, 50 HZ
Thermo Scientific Revco 11.5 Cu Ft Slimline Refrigerator, 115V.
More Details
Thermo Scientific Hamilton Airflow Products
Buy Thermo Scientific Hamilton Airflow Products Items Including:
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-19)
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-21)
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 48-well Multi (YO-01930-24).
More Details
Thermo Scientific Revco Thermo Scientific Freezers
Buy Thermo Scientific Revco Thermo Scientific Freezers Items Including:
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 17.3 c (YO-44146-25)
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-26)
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 23 cub (YO-44146-27).
More Details
Thermo Scientific Equipment
Buy Thermo Scientific Equipment Items Including:
Thermo Scientific Barnstead Pretreatment cartridge, Pore size: 10 µ (FL583X2)
Thermo Scientific Barnstead Posttreatment cartridges, Pore size: 0.2 (FL583X6)
Thermo Scientific Barnstead Posttreatment cartridges, Pore size: 0.45 (FL583X5).
More Details
Thermo Revco
Buy Thermo Revco Items Including:
Thermo Scientific Revco CxF -86°C Chest Freezer, 12.7 cubic feet, (ULT1390-10-D)
Thermo Scientific Revco CxF -86°C Chest Freezer, 12.7 cubic feet, (ULT1390-10-V)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01).
More Details
Thermo Scientific Revco Accessories
Buy Thermo Scientific Revco Accessories Items Including:
Red felt-tip replacement pen for ExF, DxF, and CxF Chart Recorder.
Remote Alarm, audible/visual, for ExF, DxF and CxF, 120V/60Hz.
Remote Alarm, audible/visual, for ExF, DxF and CxF, 230V/60Hz.
Sensaphone, 4 lines, for ExF, DxF, and CxF, 120V/60Hz..
More Details
Thermo Scientific Revco
Buy Thermo Scientific Revco Items Including:
Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-A)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 33.5 c (YO-44146-14)
Thermo Scientific Revco; ExF Series, -86°C Upright Freezer, 13 cub (YO-44146-20).
More Details
Thermo Scientific Freezer Accessories
Buy Thermo Scientific Freezer Accessories Items Including:
Accessory Key Pack for Access Control Option, for UxF.
Sliding Drawer Rack for 2" box, 20 bxs/rack, for UxF and 28 cuft ExF.
Side Access Rack for 2" box, 20 bxs/rack, fits UxF.
Sliding Drawer Rack, 20 Microplate, for UxF and 28 cuft ExF..
More Details
Thermo Scientific Revco Value Plus Freezer
Buy Thermo Scientific Revco Value Plus Freezer Items Including:
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-02)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03).
More Details
Thermo Scientific Revco Ultima Plus Freezer
Buy Thermo Scientific Revco Ultima Plus Freezer Items Including:
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-02)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03).
More Details
Thermo Scientific
Buy Thermo Scientific Items Including:
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ EasY cell cul (YO-01930-48)
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ EasY cell cul (YO-01930-49)
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ EasY cell cul (YO-01930-53).
More Details
Scientific Products
Buy Scientific Products Items Including:
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-19)
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-21)
Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-D).
More Details
Thermo Scientific Freezer Manual
Buy Thermo Scientific Freezer Manual Items Including:
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-02)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03).
More Details
Revco Freezer Accessories
Buy Revco Freezer Accessories Items Including:
6" Chart Recorder for 3 cuft Revco CxF Ultra-Low Temperature Chest Fre
6" Chart Recorder for Revco CxF Ultra-Low Temperature Chest Freezers,
2" Box Rack for Revco 3 cuft CxF Ultra-Low Temperature Chest Freezers..
More Details
70618
results
<<<Previous
26
27
28
29
30
31
32
33
34
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
44
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cole Beaker Positioning Covers to Revco Freezer Accessories
>
31125329-s-toc_2 31125329stoc2