Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 40
Adam Equipment High-Capacity Precision Toploading Balances
to
Fume Hood Shelves
Image
Product Detail
More Detail
Adam Equipment High-Capacity Precision Toploading Balances
Buy Adam Equipment High-Capacity Precision Toploading Balances Items Including:
Adam High-Capacity Precision Toploading Balance, 200 G X 0.001
Adam High-Capacity Precision Toploading Balance, 2 Kg X 0.01 G
Adam High-Capacity Precision Toploading Balance, 200 G X 0.001 G (220.
More Details
Adam Eclipse Toploading Balances
Buy Adam Eclipse Toploading Balances Items Including:
Adam PGD 253i Precision Toploader Balance, 250g w/1mg readability, gra (YO-11121-36)
Adam PGD 253i Precision Toploader Balance, 250g w/1mg readability, gra (YO-11121-37).
More Details
Adam Equipment Balances Analytical
Buy Adam Equipment Balances Analytical Items Including:
Analytical Balance Dust Cover for Adam Equipment balances (8003)
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-98)
Adam PGD 84e Analytical Balance, 80g with 0.1mg readability, graphic L (YO-11120-99).
More Details
Precision Adam Equipment Toploading Balances
Buy Precision Adam Equipment Toploading Balances Items Including:
Adam PGW Precision Toploading Balance, 150 g x 0.001 g - 115 VAC
Adam PGD 453i Precision Toploader Balance, 450g w/1mg readability, gra (YO-11121-38)
Adam PGD 453i Precision Toploader Balance, 450g w/1mg readability, gra (YO-11121-39).
More Details
Adam High-Capacity Precision Toploading Balances
Buy Adam High-Capacity Precision Toploading Balances Items Including:
Adam High-Capacity Precision Toploading Balance, 200 G X 0.001
Adam High-Capacity Precision Toploading Balance, 2 Kg X 0.01 G
Adam High-Capacity Precision Toploading Balance, 200 G X 0.001 G (220.
More Details
Adam Pgw Precision Toploading Balances
Buy Adam Pgw Precision Toploading Balances Items Including:
Adam PGW Precision Toploading Balance, 150 g x 0.001 g - 115 VAC
Adam PGW Precision Toploading Balance, 250 g x 0.001 g - 115 VAC
Adam PGW Precision Toploading Balance, 4500 g x 0.01 g - Internal Cali.
More Details
Adam Toploading Balances
Buy Adam Toploading Balances Items Including:
Adam PGW Precision Toploading Balance, 150 g x 0.001 g - 115 VAC
Adam PGD 1003e Precision Toploader Balance, 1000g with 1mg readability (YO-11121-18)
Adam PGD 1003e Precision Toploader Balance, 1000g with 1mg readability (YO-11121-19).
More Details
High Precision Balance
Buy High Precision Balance Items Including:
Adam High-Capacity Precision Toploading Balance, 200 G X 0.001 G (220
Adam High-Capacity Precision Toploading Balance, 4 Kg X 0.01 G (220 VA
Adjustable Height Mobile Base Stand, 33-40" High (5235500).
More Details
Adam Pgd 304e
Buy Adam Pgd 304e Items Including:
Adam PGD 304e Analytical Balance, 300g with 0.1mg readability, graphic (YO-11121-06).
More Details
High Precision Scales
Buy High Precision Scales Items Including:
Atago High-Precision Abbe refractometer; 0 to 95.0% Brix, 1.3000 to 1.
Adam High-Capacity Precision Toploading Balance, 200 G X 0.001
Adam High-Capacity Precision Toploading Balance, 200 G X 0.001 G (220.
More Details
Cole-Parmer Glass Tubing
Buy Cole-Parmer Glass Tubing Items Including:
Ace Glass Tube Adapter, 24/40 Joint (5028-30)
Flowtubes for carbon dioxide, 55 mL/min, glass float (YO-32048-08)
Round bottom tubes with screw-cap; 100-mL; glass; 1/case (8422-100)
Round bottom tubes with screw-cap; 50-mL; glass; 12/case (8422-50).
More Details
Glass Laboratory Tubing
Buy Glass Laboratory Tubing Items Including:
Ace Glass Tube Adapter, 24/40 Joint (5028-30)
Glass Tubing, 3mm (YO-34742-00)
Glass Tubing, 4mm (YO-34742-02)
Glass Tubing, 5mm (YO-34742-04)
Glass Tubing, 6mm (YO-34742-06)
More Details
Cole-Parmer Ace Glass Tube Adapter
Buy Cole-Parmer Ace Glass Tube Adapter Items Including:
Ace Glass Tube Adapter with Hose Barb, 24/40 Joint (5261-16).
More Details
Ace Glass Tube Adapter
Buy Ace Glass Tube Adapter Items Including:
Ace Glass Tube Adapter with Hose Barb, 24/40 Joint (5261-16).
More Details
Glass to Tubing Adapter
Buy Glass to Tubing Adapter Items Including:
Ace Glass Tube Adapter with Hose Barb, 24/40 Joint (5261-16).
More Details
Glass Tubing Sizes
Buy Glass Tubing Sizes Items Including:
Ace Glass Tube Adapter, 24/40 Joint (5028-30)
Ace Glass Tube Adapter with Hose Barb, 24/40 Joint (5261-16).
More Details
Cole Ace Glass Tube Adapter
Buy Cole Ace Glass Tube Adapter Items Including:
Ace Glass Tube Adapter with Hose Barb, 24/40 Joint (5261-16).
More Details
Cole Glass Tubing
Buy Cole Glass Tubing Items Including:
Ace Glass Tube Adapter, 24/40 Joint (5028-30)
Glass Tubing, 22mm (YO-34742-36)
Glass Tubing, 25mm (YO-34742-38)
Glass Tubing, 28mm (YO-34742-40)
Glass Tubing, 30mm (YO-34742-42)
More Details
Glass Joints
Buy Glass Joints Items Including:
Cold trap, glass, 40 mL, 29/42 joint size (7729-28)
West condenser, medium length, 190 mm, 14/20 joint (YO-34705-94)
West condenser, full length, 242 mm, 24/40 joint (YO-34705-98)
West condenser, full length, 342 mm, 24/40 joint (YO-34706-00).
More Details
Cole-Parmer Aces
Buy Cole-Parmer Aces Items Including:
Cole-Parmer Built-In Glass Prefilters with Final Sterile Cellulose Ace
Ace Glass Tube Adapter, 24/40 Joint (5028-30).
More Details
Scienceware Adapters and Stoppers
Buy Scienceware Adapters and Stoppers Items Including:
Scienceware® stopper of PTFE, size 14/20 (YO-06133-20)
Scienceware stirrer bearing of PTFE
Scienceware thermometer adapter of PTFE, size 14/20
Scienceware thermometer adapter of PTFE, size 10/18
Scienceware thermometer adapter of PTFE, size 19/22.
More Details
Scienceware Stoppers
Buy Scienceware Stoppers Items Including:
Scienceware® stopper of PTFE, size 14/20 (YO-06133-20)
Scienceware stirrer bearing of PTFE
Scienceware thermometer adapter of PTFE, size 14/20
Scienceware thermometer adapter of PTFE, size 10/18
Scienceware thermometer adapter of PTFE, size 19/22.
More Details
Scienceware Lab Supplies & Consumables
Buy Scienceware Lab Supplies & Consumables Items Including:
Scienceware® radioimmunoassay tube rack for 10- to 13-mm tubes (YO-06747-10)
Drying Rack, Benchtop, single-sided 2 tier (YO-06733-93)
Drying Rack, Benchtop, single-sided 3 tier (YO-06733-94)
Drying Rack, Benchtop, double-sided 1 tier (YO-06733-95).
More Details
Quick Connect Faucet Adapter
Buy Quick Connect Faucet Adapter Items Including:
Push-to-connect male adapter, polypropylene, 1/4" x 1/8" NPTM, 10/pack (YO-34007-42)
Push-to-connect male adapter, polypropylene, 1/4" x 1/4" NPTM, 10/pack (YO-34007-43)
Sani-Lock 3/4" Butt Weld Sanitary Adapter, 1 ea.
More Details
Quick Connect Luer Adapters
Buy Quick Connect Luer Adapters Items Including:
Push-to-connect male adapter, polypropylene, 1/4" x 1/8" NPTM, 10/pack (YO-34007-42)
Push-to-connect male adapter, polypropylene, 1/4" x 1/4" NPTM, 10/pack (YO-34007-43)
Sani-Lock 3/4" Butt Weld Sanitary Adapter, 1 ea.
More Details
Inch Push Fittings Elbow
Buy Inch Push Fittings Elbow Items Including:
Push-to-connect stem elbow adapters, 1/4" x 1/4", 10/pack (YO-34006-76)
Push-to-connect stem elbow adapters, 3/16" x 3/16", 10/pack (YO-34006-99)
Push-to-connect stem elbow adapters, 3/8" x 3/16", 10/pack (YO-34007-01).
More Details
Quick Connect Elbow
Buy Quick Connect Elbow Items Including:
Push-to-connect elbow adapters, polypropylene, 1/4", 10/pack (YO-34007-58)
Push-to-connect elbow adapters, polypropylene, 3/8" x 1/4", 10/pack (YO-34007-59)
Push-to-connect union elbow, 316 stainless steel, 1/4" (YO-31370-19).
More Details
Adapters for John Guest Polypropylene Fittings
Buy Adapters for John Guest Polypropylene Fittings Items Including:
Push-to-connect male adapter, polypropylene, 1/4" x 1/8" NPTM, 10/pack (YO-34007-42)
Push-to-connect male adapter, polypropylene, 1/4" x 1/4" NPTM, 10/pack (YO-34007-43).
More Details
Push to Connect Swivel 1 8
Buy Push to Connect Swivel 1 8 Items Including:
Push-to-connect male threaded stem adapter, 1/4" x 1/8" NPTM, 10/pack (YO-34006-66)
Push-to-connect male threaded stem adapter, 3/8" x 1/4" NPTM, 10/pack (YO-34006-68).
More Details
Fume Hood Shelves
Buy Fume Hood Shelves Items Including:
Add-on shelving unit for four-shelf starter unit 47524-01 (A1836CP64A-4)
Add-on shelving unit for four-shelf starter unit 47524-02 (A1848CP64A-4)
Add-on shelving unit for four-shelf starter unit 47524-03 (A1860CP64A-4).
More Details
70618
results
<<<Previous
36
37
38
39
40
41
42
43
44
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
53
54
60
70
80
90
100
110
120
130
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Adam Equipment High-Capacity Precision Toploading Balances to Fume Hood Shelves
>
31125329-s-toc_2 31125329stoc2