Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 400
30x Microscope
to
Cole-Parmer Symmetry Portable Balances
Image
Product Detail
More Detail
30x Microscope
Buy 30x Microscope Items Including:
Full Lens Clip-On Magnifier, 3.0x (CMG-3.0X)
Economy compact dual-power stereomicroscopes; magnification, 10x/30x; (YO-03905-10)
Leica Zoom 2000™ Stereozoom Microscope, 10x to 30x Magnification, L
Clip-On Magnifier, Large frame, 3.0x magnification.
More Details
Compact Stereos
Buy Compact Stereos Items Including:
Economy compact dual-power stereomicroscopes; magnification, 10x/30x; (YO-03905-10).
More Details
R&D E Microscopes & Magnification
Buy R&D E Microscopes & Magnification Items Including:
Binocular stereozoom microscopes; magnification from 10x to 30x; fixed (YO-03905-20)
Meiji Metallurgical Microscope; magnification, 50x; binocular, 30W hal (MX7000)
Meiji Metallurgical Microscope; magnification, 50x; trinocular, 30W ha (MX7100).
More Details
Full Size Stereo Microscopes
Buy Full Size Stereo Microscopes Items Including:
Full-Size, Dual-Power Stereomicroscope; magnification, 10x/30x; top an (YO-48920-30)
Full-Size, Dual-Power Stereomicroscope; magnification, 10x/30x; top an (YO-48920-35).
More Details
Stereo Trinocular
Buy Stereo Trinocular Items Including:
Trinocular stereozoom microscope; magnification from 10x to 40x; stand (YO-48920-20)
Stereozoom Microscopes with Fluorescent Light, Trinocular, 10x to 44x,
Stereozoom microscope system with photo tube, 7x to 45x continuous mag (EMZ-5TR + PBH+MA502).
More Details
Lab P Microscopes & Magnification
Buy Lab P Microscopes & Magnification Items Including:
Binocular stereozoom microscopes; magnification from 10x to 30x; fixed (YO-03905-20)
Meiji Metallurgical Microscope; magnification, 50x; binocular, 30W hal (MX7000)
Meiji Metallurgical Microscope; magnification, 50x; trinocular, 30W ha (MX7100).
More Details
Size 10x
Buy Size 10x Items Including:
Full-Size, Dual-Power Stereomicroscope; magnification, 10x/30x; top an (YO-48920-30).
More Details
Analytical Scales
Buy Analytical Scales Items Including:
Ohaus Pioneer Analytical Balance, 65 G X 0.1 Mg (115 VAC)
Ohaus Pioneer Analytical Balance, 110 G X 0.1 Mg (115 VAC)
Ohaus Pioneer Analytical Balance, 210 G X 0.1 Mg (115 VAC)
Ohaus Adventurer™ Pro Analytical Balance, 65 g x 0.0001 g, 115VAC, (in.
More Details
Cole-Parmer Symmetry Pa Analytical Balances
Buy Cole-Parmer Symmetry Pa Analytical Balances Items Including:
Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 115 VAC
Cole-Parmer Symmetry PA-Analytical Balance, 220g x 0.0001g -- 115 VAC
Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 220 VAC.
More Details
Cole-Parmer Symmetry Analytical Balances
Buy Cole-Parmer Symmetry Analytical Balances Items Including:
Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 115 VAC
Cole-Parmer Symmetry PA-Analytical Balance, 220g x 0.0001g -- 115 VAC
Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 220 VAC.
More Details
Symmetry Analytical Balances
Buy Symmetry Analytical Balances Items Including:
Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 115 VAC
Cole-Parmer Symmetry PA-Analytical Balance, 220g x 0.0001g -- 115 VAC
Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 220 VAC.
More Details
Analytical Balance Scales
Buy Analytical Balance Scales Items Including:
Cole-Parmer Symmetry PA-Analytical Balance, 120g x 0.0001g -- 115 VAC
Sartorius Extend™ ED Analytical Balance, 120 g x 0.1 mg
Sartorius CPA Analytical Balance, 64g X 0.1mg
Sartorius Cubis Analytical Balance, 120 G X 0.1 Mg, 120VAC (MSA124S-000-DU).
More Details
A&D W Analytical Balances
Buy A&D W Analytical Balances Items Including:
Ohaus Pioneer Analytical Balance, 65 G X 0.1 Mg (115 VAC)
Ohaus Pioneer Analytical Balance, 110 G X 0.1 Mg (115 VAC)
Ohaus Pioneer Analytical Balance, 210 G X 0.1 Mg (115 VAC)
Ohaus Adventurer™ Pro Analytical Balance, 65 g x 0.0001 g, 115VAC, (in.
More Details
Symmetry Scales
Buy Symmetry Scales Items Including:
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 115V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 115V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 230V
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 3 Kg X 0.1 G 1.
More Details
Counting Balances
Buy Counting Balances Items Including:
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 115V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 115V
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 230V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 230V.
More Details
Counting Scales
Buy Counting Scales Items Including:
Cole-Parmer Symmetry CS Counting Scale, 3 Kg X 0.1g 115V
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 115V
A&D FC-i Counting Scale, 20,000g capacity
A&D FC-i Counting Scale, 50,000g capacity
A&D HC-i Counting Scale, 3 kg/6 lb.
More Details
Cole-Parmer Symmetry Cs Counting Scales
Buy Cole-Parmer Symmetry Cs Counting Scales Items Including:
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 115V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 115V
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 230V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 230V.
More Details
Cole-Parmer Symmetry Counting Scales
Buy Cole-Parmer Symmetry Counting Scales Items Including:
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 115V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 115V
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 230V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 230V.
More Details
Symmetry Counting Scales
Buy Symmetry Counting Scales Items Including:
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 115V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 115V
Cole-Parmer Symmetry CS Counting Scale, 6 Kg X 0.2 G 230V
Cole-Parmer Symmetry CS Counting Scale, 15 Kg X 0.5 G 230V.
More Details
Cole-Parmer Symmetry Industrial Compact Sales
Buy Cole-Parmer Symmetry Industrial Compact Sales Items Including:
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 3 Kg X 0.1 G 1
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-53)
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-57).
More Details
Symmetry Industrial Compact Scales
Buy Symmetry Industrial Compact Scales Items Including:
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 3 Kg X 0.1 G 1
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-53)
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-57).
More Details
Cole-Parmer Symmetry is Compact Bench Scale
Buy Cole-Parmer Symmetry is Compact Bench Scale Items Including:
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 3 Kg X 0.1 G 1
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-53)
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-57).
More Details
Symmetry Balances Industrial and Scales
Buy Symmetry Balances Industrial and Scales Items Including:
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 3 Kg X 0.1 G 1
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-53)
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-57).
More Details
Cole-Parmer Balances Industrial and Scales
Buy Cole-Parmer Balances Industrial and Scales Items Including:
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 3 Kg X 0.1 G 1
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-53)
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-57).
More Details
Scales Sale
Buy Scales Sale Items Including:
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 3 Kg X 0.1 G 1
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-53)
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G (YO-10000-57).
More Details
Symmetry Balances & Scales
Buy Symmetry Balances & Scales Items Including:
Cole-Parmer Symmetry PR-Precision Toploading Balance, 210 g x 0.001g - (YO-10000-31)
Cole-Parmer Symmetry PR-Precision Toploading Balance, 410 g x 0.001g - (YO-10000-32)
Cole-Parmer Symmetry PR-Precision Toploading Balance, 4200 g x 0.01g -.
More Details
Symmetry Balances
Buy Symmetry Balances Items Including:
Cole-Parmer Symmetry PR-Precision Toploading Balance, 210 g x 0.001g - (YO-10000-11)
Cole-Parmer Symmetry PR-Precision Toploading Balance, 410 g x 0.001g - (YO-10000-12)
Cole-Parmer Symmetry PR-Precision Toploading Balance, 4200 g x 0.01g,.
More Details
Balance 0 01g
Buy Balance 0 01g Items Including:
Cole-Parmer Symmetry ED Portable Balance, 300g x 0.1g, 115VAC
Cole-Parmer Symmetry ED Portable Balance, 500g x 0.1g, 115VAC
Cole-Parmer Symmetry ED Portable Balance, 300g x 0.1g, 220VAC
Cole-Parmer Symmetry ED Portable Balance, 500g x 0.1g, 220VAC.
More Details
Cole-Parmer Symmetry Ed Portable Balances
Buy Cole-Parmer Symmetry Ed Portable Balances Items Including:
Cole-Parmer Symmetry ED Portable Balance, 300g x 0.1g, 115VAC
Cole-Parmer Symmetry ED Portable Balance, 500g x 0.1g, 115VAC
Cole-Parmer Symmetry ED Portable Balance, 300g x 0.1g, 220VAC
Cole-Parmer Symmetry ED Portable Balance, 500g x 0.1g, 220VAC.
More Details
Cole-Parmer Symmetry Portable Balances
Buy Cole-Parmer Symmetry Portable Balances Items Including:
Cole-Parmer Symmetry ED Portable Balance, 300g x 0.1g, 115VAC
Cole-Parmer Symmetry ED Portable Balance, 500g x 0.1g, 115VAC
Cole-Parmer Symmetry ED Portable Balance, 300g x 0.1g, 220VAC
Cole-Parmer Symmetry ED Portable Balance, 500g x 0.1g, 220VAC.
More Details
70618
results
<<<Previous
396
397
398
399
400
401
402
403
404
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
310
320
330
340
350
360
370
380
385
386
387
388
389
390
391
392
393
394
395
396
397
398
399
400
401
402
403
404
405
406
407
408
409
410
411
412
413
414
420
430
440
450
460
470
480
490
500
600
700
800
900
1,000
1,100
1,200
1,300
2,000
2,354
30x Microscope to Cole-Parmer Symmetry Portable Balances
>
31125329-s-toc_2 31125329stoc2