Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 50
Glass Fiber Filters
to
Sanitary Stainless Equipment
Image
Product Detail
More Detail
Glass Fiber Filters
Buy Glass Fiber Filters Items Including:
Glass Fiber Filters; 55mm dia; pore size, 1.6µ; thickness, 0.22mm; (GA5555MM)
Advantec Grade GA55 Glass Fiber Filters; 100/box
Advantec Grade GA200 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade GA200 Glass Fiber Filters; 142mm dia; 50/box.
More Details
Advantec Grade Ga200 Glass Fiber Filters
Buy Advantec Grade Ga200 Glass Fiber Filters Items Including:
Advantec Grade GA200 Glass Fiber Filters; 25mm dia; 50/box
Advantec Grade GA200 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade GA200 Glass Fiber Filters; 142mm dia; 50/box
Advantec Grade GA200 Glass Fiber Filters; 293mm dia; 50/box.
More Details
Advantec Grade Gf75 Glass Fiber Filters
Buy Advantec Grade Gf75 Glass Fiber Filters Items Including:
Advantec Grade GF75 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GF75 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Glass Fiber Filter Sheets
Buy Advantec Glass Fiber Filter Sheets Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Advantec 47 Mm Glass Fiber Filters
Buy Advantec 47 Mm Glass Fiber Filters Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Glass Fiber Capsule Filter
Buy Glass Fiber Capsule Filter Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Advantec Grade Gs25 Glass Fiber Filters
Buy Advantec Grade Gs25 Glass Fiber Filters Items Including:
Advantec Grade GS25 Glass Fiber Filters; 293mm dia; 50/box
Advantec Grade GS25 Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GS25 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GS25 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Advantec Grade Ga100 Glass Fiber Filters
Buy Advantec Grade Ga100 Glass Fiber Filters Items Including:
Advantec Grade GA100 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GA100 Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GA100 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GA100 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Grade Gb140 Glass Fiber Filters
Buy Advantec Grade Gb140 Glass Fiber Filters Items Including:
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Grade Ga55 Glass Fiber Filters
Buy Advantec Grade Ga55 Glass Fiber Filters Items Including:
Advantec Grade GA55 Glass Fiber Filters, 47mm dia; 100/box
Advantec Grade GA55 Glass Fiber Filters; 100/box
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GA55 Glass Fiber Filters, 25mm dia; 100/box.
More Details
Advantec Grade Dp70 Glass Fiber Filters
Buy Advantec Grade Dp70 Glass Fiber Filters Items Including:
Advantec Grade DP70 Glass Fiber Filters; 47mm dia; 50/box
Advantec Grade DP70 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade DP70 Glass Fiber Filters; 142mm dia; 50/box
Advantec Grade DP70 Glass Fiber Filters; 293mm dia; 50/box.
More Details
Advantec Grade Gd120 Glass Fiber Filters
Buy Advantec Grade Gd120 Glass Fiber Filters Items Including:
Advantec Grade GD120 Glass Fiber Filters; 47mm dia; 50/box
Advantec Grade GD120 Glass Fiber Filters; 90mm dia; 50/box
Advantec Grade GD120 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade GD120 Glass Fiber Filters; 293mm dia; 50/box.
More Details
Advantec Grade Gc50 Glass Fiber Filters
Buy Advantec Grade Gc50 Glass Fiber Filters Items Including:
Advantec Grade GC50 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GC50 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GC50 Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GC50 Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Grade Gb100r Glass Fiber Filters
Buy Advantec Grade Gb100r Glass Fiber Filters Items Including:
Advantec Grade GB100R Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB100R Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GB100R Glass Fiber Filters; 125mm dia; 100/box
Advantec Grade GB100R Glass Fiber Filters; 142mm dia; 100/box.
More Details
Advantec Glas Fiber Filters
Buy Advantec Glas Fiber Filters Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Removable Glass Fiber Filter Funnel
Buy Removable Glass Fiber Filter Funnel Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Glass Fiber Filter Sheets
Buy Glass Fiber Filter Sheets Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
47 Mm Glass Fiber Filters
Buy 47 Mm Glass Fiber Filters Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Drain Fiber Filter Paper
Buy Drain Fiber Filter Paper Items Including:
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 47mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 90mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Fiber Filter
Buy Fiber Filter Items Including:
Glass Fiber Filters; 55mm dia; pore size, 1.6µ; thickness, 0.22mm; (GA5555MM)
Advantec Grade GA55 Glass Fiber Filters; 100/box
Advantec Grade GA55 Glass Fiber Filters, 125mm dia; 100/box
Advantec Grade GB140 Glass Fiber Filters; 125mm dia; 100/box.
More Details
Ebbco W80 50 Filter Paper
Buy Ebbco W80 50 Filter Paper Items Including:
Advantec Grade No
101 Qualitative Filter Paper; 12.5 cm dia.; pack of
Advantec Grade GD120 Glass Fiber Filters; 47mm dia; 50/box
Advantec Grade GD120 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade DP70 Glass Fiber Filters; 47mm dia; 50/box.
More Details
50 Hardednd Filter Paper
Buy 50 Hardednd Filter Paper Items Including:
Advantec Grade No
101 Qualitative Filter Paper; 12.5 cm dia.; pack of
Advantec Grade GD120 Glass Fiber Filters; 47mm dia; 50/box
Advantec Grade GD120 Glass Fiber Filters; 125mm dia; 50/box
Advantec Grade DP70 Glass Fiber Filters; 47mm dia; 50/box.
More Details
25mm Paper Filter
Buy 25mm Paper Filter Items Including:
Advantec Grade GA200 Glass Fiber Filters; 25mm dia; 50/box
Advantec Grade GB140 Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GB100R Glass Fiber Filters; 25mm dia; 100/box
Advantec Grade GA100 Glass Fiber Filters; 25mm dia; 100/box.
More Details
Laboratory Pressure Filters
Buy Laboratory Pressure Filters Items Including:
Double open end polypropylene Slim-Line™ housing, 20", with pressur (158204)
Advantec Pressure Filtration Holder, for 90 mm filter; 304 Stainless S
Advantec Pressure Filtration Holder for 90 mm filters; 316 Stainless S.
More Details
Advantec Stainless Steel Syringe Filter Holders
Buy Advantec Stainless Steel Syringe Filter Holders Items Including:
Advantec Stainless Steel Filter Holder, Syringe; 13mm dia
Advantec Stainless Steel Filter Holder, Syringe; 25mm dia
Advantec Stainless Steel Filter Holder, In-Line; 25mm dia
Advantec Sanitary Stainless Steel Pressure Filtration Holder, for 90 m.
More Details
Advantec Stainless Steel Pressure Filtration Holders
Buy Advantec Stainless Steel Pressure Filtration Holders Items Including:
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-32)
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-54)
Advantec Sanitary Stainless Steel Pressure Filtration Holder, for 90 m.
More Details
Advantec Stainless Steel Holder with Reservoirs
Buy Advantec Stainless Steel Holder with Reservoirs Items Including:
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-32)
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-54).
More Details
Sanitary Stainless Steel Holders
Buy Sanitary Stainless Steel Holders Items Including:
Advantec Sanitary Stainless Steel Pressure Filtration Holder, for 90 m
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-32)
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-54).
More Details
Area Stainless Steel Hot Plate
Buy Area Stainless Steel Hot Plate Items Including:
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-32)
Advantec Stainless Steel Pressure Filtration Holder with Reservoir, fo (YO-02926-54)
Advantec Sanitary Stainless Steel Pressure Filtration Holder, for 90 m.
More Details
Sanitary Stainless Equipment
Buy Sanitary Stainless Equipment Items Including:
Advantec Sanitary Stainless Steel Pressure Filtration Holder, for 142 (YO-29900-05)
Advantec Sanitary Stainless Steel Pressure Filtration Holder, for 293 (YO-29900-10)
Sanitary In-Line Strainer Assembly; 1" Tri-Clamp; with 1/8" Perforated.
More Details
70618
results
<<<Previous
46
47
48
49
50
51
52
53
54
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
53
54
55
56
57
58
59
60
61
62
63
64
70
80
90
100
110
120
130
140
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Glass Fiber Filters to Sanitary Stainless Equipment
>
31125329-s-toc_2 31125329stoc2