Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 500
Cryogenic Tubes
to
30 Oz Polypropylene Vial Color
Image
Product Detail
More Detail
Cryogenic Tubes
Buy Cryogenic Tubes Items Including:
Thermo Scientific Nunc CryoTube cryogenic tube with conical bottom; 1.
Thermo Scientific Nunc CryoFlex™ Tubing, 15/pack
16 Place Cell Divider (for 50ml Tubes) (R3021)
Polypropylene storage boxes with unattached lid for 50 (0.5 mL) tubes, (MAKE-UP).
More Details
Plastic Flat Bottom Vial
Buy Plastic Flat Bottom Vial Items Including:
Polypropylene cryogenic vial rack, holds 50 flat bottom vials (985810)
Cyrogenic vial; 2.0 mL, external thread, flat bottom, blue-colored cap
Cyrogenic vial; 2.0 mL, external thread, flat bottom, pink-colored cap.
More Details
Cryotube Cryogenics
Buy Cryotube Cryogenics Items Including:
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (377224)
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (377267)
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (379189).
More Details
Syringe Vial 50 Ml Fermentation
Buy Syringe Vial 50 Ml Fermentation Items Including:
Syringes with PTFE luer lock; syringe capacity; 50 mL; needle gauge; 2 (YO-07939-86)
Cole-Parmer PTFE Vial, 50 mL (YO-06102-02)
Cyrogenic vial; 5.0 mL, internal thread, round bottom, natural-colored
Replacement syringe; range, 5.0 mL (YO-25700-12).
More Details
Sterile Capped Vial
Buy Sterile Capped Vial Items Including:
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- White Cap (W985863)
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Blue Cap (W985686)
100 Sterile Vials with caps in foam rack, 2-mL (YO-36270-70)
500 (Non-sterile) Vials with caps, 2-mL (YO-36270-72).
More Details
Conical Vials
Buy Conical Vials Items Including:
Thermo Scientific Nunc CryoTube cryogenic tube with conical bottom; 1..
More Details
Wheaton Cryoelite Plastic Cryogenic Vials
Buy Wheaton Cryoelite Plastic Cryogenic Vials Items Including:
Cyrogenic vial; 5.0 mL, internal thread, round bottom, natural-colored
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-26)
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-28).
More Details
Medical Supplies Vial Caps
Buy Medical Supplies Vial Caps Items Including:
Cryogenic Vial Caps, 1000/cs -- Yellow, 13mm (AKM-3100-0046)
Snap-cap sample vials, 11.1 mL (19271519-144)
Snap-cap sample vials, 18.5 mL (19272519-144)
Snap-cap sample vials, 25.9 mL (19273619-144)
Snap-cap sample vials, 37.0 mL (19275019-144).
More Details
Round Bottom Dram Vials
Buy Round Bottom Dram Vials Items Including:
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-26)
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-28)
Thermo Scientific Nunc CryoTube cryogenic tube with round bottom; 1.8 (368632).
More Details
Chrome Plastic Threaded Caps
Buy Chrome Plastic Threaded Caps Items Including:
Cyrogenic vial; 2.0 mL, external thread, flat bottom, blue-colored cap
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-26)
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-28).
More Details
Internally Threaded Tubing
Buy Internally Threaded Tubing Items Including:
Thermo Scientific Nunc CryoTube cryogenic tube with round bottom; 1.8 (368632)
Thermo Scientific Nunc CryoTube cryogenic tube with round bottom; 1.8 (363401)
Thermo Scientific Nunc CryoTube cryogenic tube with conical bottom; 1..
More Details
Wheaton Caps
Buy Wheaton Caps Items Including:
20-mL Glass Scintillation Vials; size 22 urea cap/polyetheylene disc l (986548)
20-mL Glass Scintillation Vials; size 24 urea cap/polyethylene disc li (986568)
20-mL Polyethylene Scintillation Vials; urea cap/metal foil liner, cap (986702).
More Details
Thermo Scientific Nalgene Cryogenics
Buy Thermo Scientific Nalgene Cryogenics Items Including:
Thermo Scientific Nalgene Cryogenic vial; 1.0 mL, 25/pack
Thermo Scientific Nalgene Cryogenic vial; 1.5 mL, 25/pack
Thermo Scientific Nalgene Cryogenic vial; 1.2 mL, 25/pack
Thermo Scientific Nalgene Standard Cryocane, Aluminum, 300 mL (holds 6.
More Details
Wheaton Cryogenic Vials
Buy Wheaton Cryogenic Vials Items Including:
Cryogenic Vial, Sterile 1mL, Round Bottom (W985860)
Cryogenic Vial, Sterile 2.0 mL, Round Bottom (W985861)
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Yellow Cap (W985866)
Polypropylene cryogenic vial rack, holds 50 round bottom vials (985800).
More Details
Colored Dram Vials Plastic
Buy Colored Dram Vials Plastic Items Including:
Cryogenic Vial, Sterile 2.0 mL, Round Bottom (W985861)
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- White Cap (W985863)
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Green Cap (W985867)
Savillex 4mL PFA Vials in organizer trays, 10/tray (0217T).
More Details
Flat Bottom Glass Vials
Buy Flat Bottom Glass Vials Items Including:
Cyrogenic vial; 2.0 mL, external thread, flat bottom, blue-colored cap
Cyrogenic vial; 2.0 mL, external thread, flat bottom, pink-colored cap
Cyrogenic vial; 2.0 mL, internal thread, flat bottom, white-colored ca.
More Details
Blue Vials
Buy Blue Vials Items Including:
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Blue Cap (W985686)
Thermo Scientific Nalgene Colored Cap Inserts (for vials), Blue
Open Top Aluminum Seals, Blue, 13mm, 1000/CS (224177-05)
Open Top Aluminum Seals, Blue, 20mm, 1000/CS (224178-05).
More Details
Clear Plastic Vial Round End
Buy Clear Plastic Vial Round End Items Including:
Cyrogenic vial; 5.0 mL, internal thread, round bottom, natural-colored
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-26)
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-28).
More Details
Corning External Thread Cryogenic Vials
Buy Corning External Thread Cryogenic Vials Items Including:
Corning External Thread Cryogenic Vials; 2.0 mL, Self-Standing, 50/pac.
More Details
Wheaton Cryule Plastic Cryogenic Vials
Buy Wheaton Cryule Plastic Cryogenic Vials Items Including:
Cryogenic Vial, Sterile 1mL, Round Bottom (W985860)
Cryogenic Vial, Sterile 2.0 mL, Round Bottom (W985861)
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- White Cap (W985863)
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Yellow Cap (W985866).
More Details
Nunc Vials
Buy Nunc Vials Items Including:
Thermo Scientific Nunc™ CryoTube Vial Holder
Thermo Scientific Nunc CryoFlex™ Tubing, 15/pack
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (377224)
Thermo Scientific Nunc CryoTube cryogenic tube with conical bottom; 1..
More Details
Cryotube Vials
Buy Cryotube Vials Items Including:
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (377224)
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (377267)
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (379189).
More Details
Wheaton Cryogenic Vials and Caps
Buy Wheaton Cryogenic Vials and Caps Items Including:
Cryogenic Vial, Sterile 1mL, Round Bottom (W985860)
Cryogenic Vial, Sterile 2.0 mL, Round Bottom (W985861)
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- Yellow Cap (W985866)
Cyrogenic vial; 5.0 mL, internal thread, round bottom, natural-colored.
More Details
Cole-Parmer Cryogenic Vials and Caps
Buy Cole-Parmer Cryogenic Vials and Caps Items Including:
Cryogenic Vial Caps, 1000/cs -- Green, 13mm (AKM-3100-0042).
More Details
Cryogenic Vials and Caps
Buy Cryogenic Vials and Caps Items Including:
Cryogenic Vial Caps, 1000/cs -- Green, 13mm (AKM-3100-0042)
Cryogenic Vial Caps, 1000/cs -- White, 13mm (AKM-3100-0045)
Cryogenic Vial Caps, 1000/cs -- Yellow, 13mm (AKM-3100-0046)
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- White Cap (W985863).
More Details
Laboratory Vials
Buy Laboratory Vials Items Including:
Vial Crimper (W225301)
Cryogenic Vial Caps, 1000/cs -- Green, 13mm (AKM-3100-0042)
Cryogenic Vial Caps, 1000/cs -- Yellow, 13mm (AKM-3100-0046)
Scienceware serum vial rack for 10- to 13-mm vials
Scienceware serum vial rack for 13- to 16-mm vials.
More Details
Cole-Parmer Polypropylene Cryogenic Vial Caps
Buy Cole-Parmer Polypropylene Cryogenic Vial Caps Items Including:
Cryogenic Vial Caps, 1000/cs -- Green, 13mm (AKM-3100-0042).
More Details
Polypropylene Cryogenic Vial Caps
Buy Polypropylene Cryogenic Vial Caps Items Including:
Cyrogenic vial; 5.0 mL, internal thread, round bottom, natural-colored
Cyrogenic vial; 4.0 mL, internal thread, round bottom, natural-colored
Cyrogenic vial; 2.0 mL, external thread, round bottom, natural-colored (YO-03756-26).
More Details
White Plastic Vials
Buy White Plastic Vials Items Including:
Cryogenic Vial, Sterile 2.0 mL, Flat Bottom -- White Cap (W985863)
Cryogenic Vial, non-sterile 2mL, Round Bottom without white marking ar (985726).
More Details
30 Oz Polypropylene Vial Color
Buy 30 Oz Polypropylene Vial Color Items Including:
Cryogenic Vial Caps, 1000/cs -- Green, 13mm (AKM-3100-0042)
Cryogenic Vial Caps, 1000/cs -- Red, 13mm (AKM-3100-0044)
Cryogenic Vial Caps, 1000/cs -- White, 13mm (AKM-3100-0045)
Cryogenic Vial Caps, 1000/cs -- Yellow, 13mm (AKM-3100-0046).
More Details
70618
results
<<<Previous
496
497
498
499
500
501
502
503
504
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
410
420
430
440
450
460
470
480
485
486
487
488
489
490
491
492
493
494
495
496
497
498
499
500
501
502
503
504
505
506
507
508
509
510
511
512
513
514
520
530
540
550
560
570
580
590
600
700
800
900
1,000
1,100
1,200
1,300
1,400
2,000
2,354
Cryogenic Tubes to 30 Oz Polypropylene Vial Color
>
31125329-s-toc_2 31125329stoc2