Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 60
Cole Quiet Vacuum
to
Gast Air
Image
Product Detail
More Detail
Cole Quiet Vacuum/Pressure Pumps
Buy Cole Quiet Vacuum/Pressure Pumps Items Including:
Vacuum/Pressure Pump, 2.0 CFM 115V
Quiet Pressure Pump, 2.3 CFM, 3.0psi max, 115VAC.
More Details
Vacuum Pump Extreme Quiet
Buy Vacuum Pump Extreme Quiet Items Including:
Quiet Pressure Pump, 2.3 CFM, 3.0psi max, 115VAC
Quiet Pressure Pump, 3.2 CFM, 3.0psi max, 115VAC.
More Details
Ultra-Quiet Air Compressors with Storage Tank
Buy Ultra-Quiet Air Compressors with Storage Tank Items Including:
Ultra-Quiet Oil-lubricated Air Compressor, 0.7 CFM (20 LPM), 0.93 Gall
Ultra-Quiet Oil-lubricated Air Compressor, 1.05 CFM (30 LPM), 0.93 Gal (YO-07067-13)
Ultra-Quiet Oil-lubricated Air Compressor, 1.05 CFM (30 LPM), 0.93 Gal (YO-07067-14).
More Details
Piston Compressors
Buy Piston Compressors Items Including:
Tank-Mounted Piston Compressor, 2.3 cfm
Tank-Mounted Piston Compressor, 3.5 cfm
Tank-Mounted Piston Compressor, 11.0 cfm
Tank-Mounted Piston Compressor, 22.0 cfm
Oilless Air Compressor, Piston compressor pump, 3.5 cfm, 115/230 VAC (4HCC-10-M450X).
More Details
Air Compressors Portable
Buy Air Compressors Portable Items Including:
Professional series air compressor, 7.1 cfm, 20 gallon portable tank,.
More Details
Single-Stage Air Compressors with Storage Tanks
Buy Single-Stage Air Compressors with Storage Tanks Items Including:
Professional series air compressor, 7.1 cfm, 20 gallon portable tank,
Direct drive air compressor, 3.0 cfm, hotdog tank, 115 VAC (LA5706)
Direct drive air compressor, 4.8 cfm (136 LPM), 2 gallon (7.5 liters).
More Details
Quiet Pressure Pumps
Buy Quiet Pressure Pumps Items Including:
Quiet Pressure Pump, 2.3 CFM, 3.0psi max, 115VAC
Quiet Pressure Pump, 3.2 CFM, 3.0psi max, 115VAC.
More Details
Two-Stage Air Compressors with Storage Tank
Buy Two-Stage Air Compressors with Storage Tank Items Including:
Two-stage stationary, electric- driven air compressors, 16.8 cfm
Two-stage stationary, electric- driven air compressors, 24.0 cfm
Two-stage stationary, electric- driven air compressors, 35.0 cfm
Start-up kit for compressors with 120 gallon tank.
More Details
Quiet Vacuum Pumps
Buy Quiet Vacuum Pumps Items Including:
Quiet Pressure Pump, 2.3 CFM, 3.0psi max, 115VAC
Quiet Pressure Pump, 3.2 CFM, 3.0psi max, 115VAC.
More Details
Portable Compressors
Buy Portable Compressors Items Including:
Professional series air compressor, 7.1 cfm, 20 gallon portable tank,.
More Details
Air Compressor Electric
Buy Air Compressor Electric Items Including:
Two-stage stationary, electric- driven air compressors, 16.8 cfm
Two-stage stationary, electric- driven air compressors, 24.0 cfm
Two-stage stationary, electric- driven air compressors, 35.0 cfm.
More Details
Tank-Mounted Piston Compressors
Buy Tank-Mounted Piston Compressors Items Including:
Tank-Mounted Piston Compressor, 2.3 cfm
Tank-Mounted Piston Compressor, 3.5 cfm
Tank-Mounted Piston Compressor, 9.1 cfm
Tank-Mounted Piston Compressor, 11.0 cfm.
More Details
Oil Storage Tanks
Buy Oil Storage Tanks Items Including:
Ultra-Quiet Oil-lubricated Air Compressor, 0.7 CFM (20 LPM), 0.93 Gall
Ultra-Quiet Oil-lubricated Air Compressor, 1.05 CFM (30 LPM), 0.93 Gal (YO-07067-13)
Ultra-Quiet Oil-lubricated Air Compressor, 1.05 CFM (30 LPM), 0.93 Gal (YO-07067-14).
More Details
Portable Liquid Storage Tank
Buy Portable Liquid Storage Tank Items Including:
Professional series air compressor, 7.1 cfm, 20 gallon portable tank,.
More Details
Portable Tube Air Pump
Buy Portable Tube Air Pump Items Including:
Masterflex I/P ATEX Zone 2 Air-Powered Pump System, 100-650 rpm.
High-head PP tube set for electric and air motors, 27"L
High-head PP tube set for electric and air motors, 39"L
High-head PP tube set for electric and air motors, 47"L.
More Details
Portable Air Pumps
Buy Portable Air Pumps Items Including:
Professional series air compressor, 7.1 cfm, 20 gallon portable tank,.
More Details
Air Lubricator
Buy Air Lubricator Items Including:
Ultra-Quiet Oil-lubricated Air Compressor, 4.5 cfm, 115 VAC (P150/50AL)
Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4"
Mini Air Regulator, Max 45 SCFM, Pressure range of 0 - 140 PSIG, 1/8".
More Details
Quiet Bubbles Air Pumps
Buy Quiet Bubbles Air Pumps Items Including:
Ultra-Quiet Oilless Air Compressor, 1.9 cfm, 115 VAC (PC106/115V)
Ultra-Quiet Oilless Air Compressor, 1.9 cfm, 220 VAC (PC106/220V)
Ultra-Quiet Oilless Air Compressor, 3.5 cfm, 115 VAC (PC224E/115V)
Quiet Pressure Pump, 2.3 CFM, 3.0psi max, 115VAC.
More Details
50 Gallon Tank Mixers
Buy 50 Gallon Tank Mixers Items Including:
Ultra-Quiet Oil-lubricated Air Compressor, 5.0 CFM (134 LPM), 6 Gallon (YO-07067-51).
More Details
Air Tank Kit
Buy Air Tank Kit Items Including:
Start-up kit for compressors with 80 gallon tank
Start-up kit for compressors with 120 gallon tank.
More Details
Air Tanks Portable
Buy Air Tanks Portable Items Including:
Professional series air compressor, 7.1 cfm, 20 gallon portable tank,.
More Details
Cole Ultra-Quiet Air Compressors
Buy Cole Ultra-Quiet Air Compressors Items Including:
Ultra-Quiet Oil-lubricated Air Compressor, 1.5 cfm, 115 VAC (P50/24AL)
Ultra-Quiet Oil-lubricated Air Compressor, 4.5 cfm, 115 VAC (P150/50AL)
Ultra-Quiet Oil-lubricated Air Compressor, 1.05 CFM (30 LPM), 0.93 Gal (YO-07067-13).
More Details
Air Compressor Series
Buy Air Compressor Series Items Including:
Contractor series air compressor, 6.0 cfm, pancake tank, 115 VAC (DP2004V)
Mini Air Regulator, Max 59 SCFM, Pressure range of 0 - 140 PSIG, 1/4"
Mini Air Regulator, Max 45 SCFM, Pressure range of 0 - 140 PSIG, 1/8".
More Details
Air Compressors Direct
Buy Air Compressors Direct Items Including:
Direct drive air compressor, 3.0 cfm, hotdog tank, 115 VAC (LA5706)
Direct drive air compressor, 4.8 cfm (136 LPM), 2 gallon (7.5 liters).
More Details
Electric Compressor
Buy Electric Compressor Items Including:
Two-stage stationary, electric- driven air compressors, 16.8 cfm
Two-stage stationary, electric- driven air compressors, 24.0 cfm
Two-stage stationary, electric- driven air compressors, 35.0 cfm.
More Details
Piston Air
Buy Piston Air Items Including:
Tank-Mounted Piston Compressor, 2.3 cfm
Tank-Mounted Piston Compressor, 3.5 cfm
Tank-Mounted Piston Compressor, 11.0 cfm
Tank-Mounted Piston Compressor, 22.0 cfm
Oilless Air Compressor, Piston compressor pump, 3.5 cfm, 115/230 VAC (4HCC-10-M450X).
More Details
Stage Air Compressor
Buy Stage Air Compressor Items Including:
Direct drive air compressor, 3.0 cfm, hotdog tank, 115 VAC (LA5706)
Two-stage stationary, electric- driven air compressors, 16.8 cfm
Two-stage stationary, electric- driven air compressors, 24.0 cfm
Start-up kit for compressors with 120 gallon tank.
More Details
Gast Compressor
Buy Gast Compressor Items Including:
Tank-Mounted Piston Compressor, 2.3 cfm
Tank-Mounted Piston Compressor, 3.5 cfm
Tank-Mounted Piston Compressor, 9.1 cfm
Tank-Mounted Piston Compressor, 11.0 cfm
Tank-Mounted Piston Compressor, 22.0 cfm
More Details
Tank 120
Buy Tank 120 Items Including:
Excalibur tank, 120 gallons, 1.35 specific gravity (YO-06960-18)
Refrigerated tank; 120 VAC, 60 Hz (YO-12110-00)
Start-up kit for compressors with 120 gallon tank
Ultra-Quiet Oil-lubricated Air Compressor, 4.2 CFM (120 LPM), 6.3 Gall (YO-07067-23).
More Details
Gast Air
Buy Gast Air Items Including:
Tank-Mounted Piston Compressor, 2.3 cfm
Tank-Mounted Piston Compressor, 3.5 cfm
Tank-Mounted Piston Compressor, 9.1 cfm
Tank-Mounted Piston Compressor, 11.0 cfm
Tank-Mounted Piston Compressor, 22.0 cfm
More Details
70618
results
<<<Previous
56
57
58
59
60
61
62
63
64
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
45
46
47
48
49
50
51
52
53
54
55
56
57
58
59
60
61
62
63
64
65
66
67
68
69
70
71
72
73
74
80
90
100
110
120
130
140
150
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cole Quiet Vacuum to Gast Air
>
31125329-s-toc_2 31125329stoc2