Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 7
Controls and Automation
to
30 V Ac Power Supply
Image
Product Detail
More Detail
Controls and Automation
Buy Controls and Automation Items Including:
AC Variable-Speed Controller, 1/3 hp, 115/230 VAC, 1.7 amp, 120-240VAC
AC Variable-Speed Controller, 1/2 hp, 115/230 VAC, 2.4 amp, 120-240VAC
AC Variable-Speed Controller, 1 hp, 115/230 VAC, 4.2 amp, 120-240VAC o
380 Vac25 Amp Dc Control.
More Details
Ac Motor Controllers
Buy Ac Motor Controllers Items Including:
AC Variable-Speed Controller, 1/3 hp, 115/230 VAC, 1.7 amp, 120-240VAC
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV251N02SXB)
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV371N02YXB).
More Details
Inverters Ac
Buy Inverters Ac Items Including:
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV251N02SXB)
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV371N02YXB).
More Details
Automation Control Ac Controllers
Buy Automation Control Ac Controllers Items Including:
AC Variable-Speed Controller, 1/3 hp, 115/230 VAC, 1.7 amp, 120-240VAC
AC Drive / Frequency Inverter, 480-600 VAC three phase input with 3 ph (ESV152N06TXB)
AC Drive / Frequency Inverter, 480-600 VAC three phase input with 3 ph (ESV222N06TXB).
More Details
Ac Controllers
Buy Ac Controllers Items Including:
AC Variable-Speed Controller, 1/3 hp, 115/230 VAC, 1.7 amp, 120-240VAC
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV251N02SXB)
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV371N02YXB).
More Details
Automation Control Motors
Buy Automation Control Motors Items Including:
AC Variable-Speed Controller, 1/3 hp, 115/230 VAC, 1.7 amp, 120-240VAC
AC Drive / Frequency Inverter, 480-600 VAC three phase input with 3 ph (ESV152N06TXB)
AC Drive / Frequency Inverter, 480-600 VAC three phase input with 3 ph (ESV222N06TXB).
More Details
Ac Ph Control
Buy Ac Ph Control Items Including:
AC Drive / Frequency Inverter, 200-240 VAC three phase input with 3 ph (ESV112N02TXB)
AC Drive / Frequency Inverter, 200-240 VAC three phase input with 3 ph (ESV152N02TXB).
More Details
Ac Motor Drives
Buy Ac Motor Drives Items Including:
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV251N02SXB)
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV371N02YXB).
More Details
Automation Control Pumps
Buy Automation Control Pumps Items Including:
AC Variable-Speed Controller, 1/3 hp, 115/230 VAC, 1.7 amp, 120-240VAC.
More Details
200 Inverter
Buy 200 Inverter Items Including:
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV251N02SXB)
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV371N02YXB).
More Details
Autom Ac Controllers
Buy Autom Ac Controllers Items Including:
AC Variable-Speed Controller, 1/3 hp, 115/230 VAC, 1.7 amp, 120-240VAC
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV251N02SXB)
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV371N02YXB).
More Details
High Frequency Ac Power Supplies
Buy High Frequency Ac Power Supplies Items Including:
1653A : 150V 2A AC Power Supply (1653A)
Triple Output, High-Current DC Power Supply (GPC-3030D)
High current linear power supply 32VDC - 20A, LED readout (1790)
High-Current, Single-Output DC Power Supply, 0 to 18 VDC, 0 to 10 A (GPR-1810HD).
More Details
110 Volt Ac Power Supply
Buy 110 Volt Ac Power Supply Items Including:
Power Supply/Adapter (110-00-18T)
Power Supply/Adapter (110-00-08T)
Dual-output Electrophoresis Power Supply, 0-150 volts, 100/240VAC (RS-PS-75)
24 Volt DC transmitter power supply module with connector (24-TP)
9 Volt DC Adapter 110V Power Supply.
More Details
Regulated Ac Power Supply
Buy Regulated Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
Extech Digital Regulated DC Power Supply, 115/220 VAC
B&K Precision 1657 AC Power Source, 1500 VA AC
1689 : Regulated Analog 28A DC Power Supply (1689).
More Details
110v Ac Power Supply
Buy 110v Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC.
More Details
Portable Ac Power Supply
Buy Portable Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC
AC Power Adapter 115VAC (PC5OPT01)
Power supply / charger for Drummond Portable Pipet-Aid XL model.
More Details
20 Volt Ac Power Supply
Buy 20 Volt Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC
Thermo Scientific Electrophoresis Power Supply 300v 120v (YO-28406-20)
Solid Core:3-Phase4 Wire 4-20 Ma Ac Voltage.
More Details
Constant Current Ac Power Supply
Buy Constant Current Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
Triple Output, High-Current DC Power Supply (GPC-3030D)
High current linear power supply 32VDC - 20A, LED readout (1790)
High-Current, Single-Output DC Power Supply, 0 to 18 VDC, 0 to 10 A (GPR-1810HD).
More Details
Ac Power Supply 20v Output
Buy Ac Power Supply 20v Output Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC
Quad Output DC Power Supply (382270)
High-Current, Single-Output DC Power Supply, 0 to 18 VDC, 0 to 3 A (GPS-1830D).
More Details
100 Hz Ac Power Supply
Buy 100 Hz Ac Power Supply Items Including:
B&K Precision 1657 AC Power Source, 1500 VA AC
Dual-output Electrophoresis Power Supply, 0-150 volts, 100/240VAC (RS-PS-75)
Power supply for 97606-00 and -02, 115 VAC/60 Hz (99005501)
Power supply for 97606-00 and -02, 220 VAC/50 Hz (99005502).
More Details
12 Volt Ac Power Supply
Buy 12 Volt Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC
Fixed DC power supply; 12 VDC out, 625 mA (YO-26900-11)
Tektronix PWS4721 Programmable DC Power Supply, 72 V, 1.2 A.
More Details
Ac Power Supply Blackberry Playbook
Buy Ac Power Supply Blackberry Playbook Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
4 Kv Ac Power Supply
Buy 4 Kv Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
Ovation 4-Connection Pipette Power Supply, all countries (Universal)
Single Phase 4-20MA Ac Current True Rms Transducers
Single Phase 4/20MA Ac Current Ave Rms Xducer.
More Details
Programmable Ac Power Supplies
Buy Programmable Ac Power Supplies Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision XLN3640, 1.44 kW Programmable DC Power Supply, 0 to 36 V
B&K Precision XLN3640-GL, 1.44 kW Programmable DC Power Supply, 0 to 3.
More Details
Ac Power Supplies Pzqan3 570
Buy Ac Power Supplies Pzqan3 570 Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Crane Game Ac Power Supply
Buy Crane Game Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Ac to Ac Power Supply
Buy Ac to Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Dc to Ac Power Supply
Buy Dc to Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC
Quad Output DC Power Supply (382270)
High-Current, Single-Output DC Power Supply, 0 to 18 VDC, 0 to 3 A (GPS-1830D).
More Details
Ac Power Supply Connection
Buy Ac Power Supply Connection Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC.
More Details
30 V Ac Power Supply
Buy 30 V Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
High-Current, Single-Output DC Power Supply, 0 to 30 VDC, 0 to 3 A (GPS-3030D)
Single Output Power Supply, Analog Display, 0 to 30 VDC 1626A (1626A)
Tektronix PWS4305 Programmable DC Power Supply, 30 V, 5 A.
More Details
70618
results
<<<Previous
3
4
5
6
7
8
9
10
11
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Controls and Automation to 30 V Ac Power Supply
>
31125329-s-toc_2 31125329stoc2