Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 700
Soxhlet System
to
Ce Compliance
Image
Product Detail
More Detail
Soxhlet System
Buy Soxhlet System Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet extraction glassware systems, 125 mL (YO-99036-10).
More Details
Micro Kjeldahl Apparatus
Buy Micro Kjeldahl Apparatus Items Including:
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 115V
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 230V (MM2313/E) (YO-03074-32).
More Details
Extraction Glassware
Buy Extraction Glassware Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Soxhlet extraction glassware systems, 250 mL (YO-99036-20)
Kuderna-Danish Concentrator without springs, 500mL capacity, 615mm H.
More Details
Stirrer with Heater
Buy Stirrer with Heater Items Including:
Electrothermal Electromantle Extraction Heater - 3 positions, 1000 mL, (EME3 1000/CEBX6)
Electrothermal Electromantle Extraction Heating/Stirrer - 3 positions, (EMEA3 0100/CEX1).
More Details
Soxhlet Extraction Systems
Buy Soxhlet Extraction Systems Items Including:
500 ml round bottom Soxhlet Allihn bulb condenser (YO-99037-18)
125 ml Soxhlet extractor body (YO-99037-38)
250 ml Soxhlet extractor body (YO-99037-40)
Soxhlet 125 ml flat bottom flask (YO-99037-44)
Soxhlet 125 ml round bottom flask (YO-99037-60).
More Details
Cole-Parmer Soxhlet Extraction Glassware Systems
Buy Cole-Parmer Soxhlet Extraction Glassware Systems Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Soxhlet extraction glassware systems, 250 mL (YO-99036-20).
More Details
Using a Soxhlet Extraction Apparatus
Buy Using a Soxhlet Extraction Apparatus Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet 125 ml flat bottom flask (YO-99037-44)
Soxhlet 250 ml flat bottom flask (YO-99037-46)
Soxhlet 300 ml flat bottom flask (YO-99037-48)
Soxhlet 125 ml round bottom flask (YO-99037-60).
More Details
Electrothermal Extraction Apparatus
Buy Electrothermal Extraction Apparatus Items Including:
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 115V
Electrothermal Electromantle Extraction Heater - 3 positions, 1000 mL, (EME3 1000/CEBX6)
Electrothermal Electromantle Extraction Heating/Stirrer - 3 positions, (EMEA3 0100/CEX1).
More Details
Cole-Parmer Extraction Apparatus
Buy Cole-Parmer Extraction Apparatus Items Including:
Paley bottle, cheese test, 3/case (YO-34590-40)
Babcock variable-speed Orbital Shaker;110V/60Hz, 24 bottle capacity
Babcock variable-speed Orbital Shaker;220V/50Hz, 24 bottle capacity
500 ml round bottom Soxhlet Allihn bulb condenser (YO-99037-18).
More Details
Cole Extraction
Buy Cole Extraction Items Including:
Cole-Parmer Yeast Extract, 500 g
Mojonnier extraction flask (YO-34590-65)
Buffered beef extract broth, prepared; 1 liter, 10/pk (YO-14204-52)
Gerber-method Butyrometer; milk test (YO-17425-10)
Babcock bottle, skim milk test, 12/case (YO-34590-00).
More Details
Soxhlet Extraction Glassware
Buy Soxhlet Extraction Glassware Items Including:
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Soxhlet extraction glassware systems, 250 mL (YO-99036-20)
Extraction thimble for soxhlet extraction glassware systems, 25 mL (YO-99036-50).
More Details
Soxhlet Extraction
Buy Soxhlet Extraction Items Including:
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Extraction thimble for soxhlet extraction glassware systems, 25 mL (YO-99036-50)
500 ml round bottom Soxhlet Allihn bulb condenser (YO-99037-18)
125 ml Soxhlet extractor body (YO-99037-38).
More Details
Soxhlet Extraction Apparatus Holder
Buy Soxhlet Extraction Apparatus Holder Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet 125 ml flat bottom flask (YO-99037-44)
Soxhlet 250 ml flat bottom flask (YO-99037-46)
Soxhlet 300 ml flat bottom flask (YO-99037-48)
Soxhlet 125 ml round bottom flask (YO-99037-60).
More Details
Electrothermal Mm Series Micro-Kjeldahl Extraction Heater
Buy Electrothermal Mm Series Micro-Kjeldahl Extraction Heater Items Including:
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 115V
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 230V (MM2313/E) (YO-03074-32).
More Details
Electrothermal Mq Series Macro-Kjeldahl Extraction Heaters
Buy Electrothermal Mq Series Macro-Kjeldahl Extraction Heaters Items Including:
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 30 (MQ3822B/E)
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 80 (MQ3824B/E)
Electrothermal MQ Series Six-well Macro-Kjeldahl Extraction Heater, 80 (MQ3868B/E).
More Details
Cole-Parmer Soxhlet Extraction Systems
Buy Cole-Parmer Soxhlet Extraction Systems Items Including:
Soxhlet extraction glassware systems, 125 mL (YO-99036-10).
More Details
Soxhlet Glassware
Buy Soxhlet Glassware Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Soxhlet extraction glassware systems, 250 mL (YO-99036-20)
500 ml round bottom Soxhlet Allihn bulb condenser (YO-99037-18).
More Details
Programmable Soxhlet Extraction Systems
Buy Programmable Soxhlet Extraction Systems Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet 125 ml flat bottom flask (YO-99037-44)
Soxhlet 250 ml flat bottom flask (YO-99037-46)
Soxhlet 300 ml flat bottom flask (YO-99037-48)
Soxhlet 125 ml round bottom flask (YO-99037-60).
More Details
Automated Soxhlet Extraction System
Buy Automated Soxhlet Extraction System Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet 125 ml flat bottom flask (YO-99037-44)
Soxhlet 250 ml flat bottom flask (YO-99037-46)
Soxhlet 300 ml flat bottom flask (YO-99037-48)
Soxhlet 125 ml round bottom flask (YO-99037-60).
More Details
Extraction Heating/Stirrer
Buy Extraction Heating/Stirrer Items Including:
Electrothermal Electromantle Extraction Heating/Stirrer - 3 positions, (EMEA3 0100/CEX1)
Electrothermal Electromantle Extraction Heating/Stirrer - 3 positions, (EMEA3 0100/CEX6).
More Details
Kjeldahl Equipment Extraction Heaters
Buy Kjeldahl Equipment Extraction Heaters Items Including:
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 115V
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 30 (MQ3822B/EX1)
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 30 (MQ3822B/EX6).
More Details
Cole Soxhlet Extraction Glassware Systems
Buy Cole Soxhlet Extraction Glassware Systems Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Soxhlet extraction glassware systems, 250 mL (YO-99036-20).
More Details
Extraction Heater
Buy Extraction Heater Items Including:
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 115V
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 30 (MQ3822B/EX1)
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 30 (MQ3822B/EX6).
More Details
Extraction Systems
Buy Extraction Systems Items Including:
Extract-All Portable Air Cleaning System with 4' to 6' Long Extractor
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Secondary Merv 8 Pleated Filter
Primary Merv 14 Cell Filter.
More Details
Cole Soxhlet Extraction Systems
Buy Cole Soxhlet Extraction Systems Items Including:
500 ml round bottom Soxhlet Allihn bulb condenser (YO-99037-18)
125 ml Soxhlet extractor body (YO-99037-38)
250 ml Soxhlet extractor body (YO-99037-40)
Soxhlet 125 ml flat bottom flask (YO-99037-44)
Soxhlet 125 ml round bottom flask (YO-99037-60).
More Details
Electrothermal Lab Equipment
Buy Electrothermal Lab Equipment Items Including:
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 115V
Electrothermal Electromantle Extraction Heater - 3 positions, 100 mL, (EME3 0100/CEBX1)
Electrothermal Electromantle Extraction Heater - 3 positions, 1000 mL, (EME3 1000/CEBX6).
More Details
Electrothermal Extraction
Buy Electrothermal Extraction Items Including:
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 115V
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 30 (MQ3822B/EX1)
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 30 (MQ3822B/EX6).
More Details
Cole-Parmer Extraction
Buy Cole-Parmer Extraction Items Including:
Cole-Parmer Yeast Extract, 500 g
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Soxhlet extraction glassware systems, 250 mL (YO-99036-20)
Mojonnier extraction flask (YO-34590-65)
C18, water wettable SPE cartridges, 3 mL (YO-10937-06).
More Details
Cole Extraction Apparatus
Buy Cole Extraction Apparatus Items Including:
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Mojonnier extraction flask (YO-34590-65)
Babcock bottle, skim milk test, 12/case (YO-34590-00)
Babcock bottle, skim milk test (YO-34590-01)
Babcock bottle, milk test, 12/case (YO-34590-05).
More Details
Ce Compliance
Buy Ce Compliance Items Including:
GE (Druck) DPI 620 CE Multifunction Calibrator with Windows CE (Now ca
Electrothermal Electromantle Extraction Heating/Stirrer - 3 positions, (EMEA3 0100/CE)
Electrothermal Electromantle Extraction Heating/Stirrer - 3 positions, (EMEA3 0250/CE).
More Details
70618
results
<<<Previous
696
697
698
699
700
701
702
703
704
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
610
620
630
640
650
660
670
680
685
686
687
688
689
690
691
692
693
694
695
696
697
698
699
700
701
702
703
704
705
706
707
708
709
710
711
712
713
714
720
730
740
750
760
770
780
790
800
900
1,000
1,100
1,200
1,300
1,400
1,500
1,600
2,000
2,354
Soxhlet System to Ce Compliance
>
31125329-s-toc_2 31125329stoc2