Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 8
Variable Ac Power Supplies
to
1 Rpm Ac Motor
Image
Product Detail
More Detail
Variable Ac Power Supplies
Buy Variable Ac Power Supplies Items Including:
1653A : 150V 2A AC Power Supply (1653A).
More Details
P1350 Ac Power Supply
Buy P1350 Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Ac Power Supplies
Buy Ac Power Supplies Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Power Failure Ac Power Supplies
Buy Power Failure Ac Power Supplies Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Industrial Ac Power Supplies
Buy Industrial Ac Power Supplies Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Allen Bradley Ac Power Supply
Buy Allen Bradley Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Garmin Nuvi Ac Power Supply
Buy Garmin Nuvi Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Isolated Ac Power Supply
Buy Isolated Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Korg Ka173 Ac Power Supply
Buy Korg Ka173 Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Sdr H40p Ac Power Supply
Buy Sdr H40p Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
15v 1100ma Ac Power Supply
Buy 15v 1100ma Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Yamaha Ac Power Supply
Buy Yamaha Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Maxim Ac Power Supply
Buy Maxim Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
6000l M Ac Power Supply
Buy 6000l M Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Ac Power Supply Acuo38a
Buy Ac Power Supply Acuo38a Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Slide Ac Power Supply
Buy Slide Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
24volt Ac Power Supply
Buy 24volt Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Apple Ac Power Supply
Buy Apple Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Ac Power Supply 035a
Buy Ac Power Supply 035a Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Ac Power Supply 400hz
Buy Ac Power Supply 400hz Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Jvc Ac Power Supply
Buy Jvc Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
Rca Phone Ac Power Supply
Buy Rca Phone Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A).
More Details
90w Ac Power Supply
Buy 90w Ac Power Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC.
More Details
Ac Power Supply Systems
Buy Ac Power Supply Systems Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
B&K Precision 1657 AC Power Source, 1500 VA AC
Power Supply/Adaptor
120 VAC (110-00-08)
More Details
Ac Supply
Buy Ac Supply Items Including:
1653A : 150V 2A AC Power Supply (1653A)
1655A : 150V 3A AC Power Supply (1655A)
BMP71 AC/Battery adapter
BMP21 AC Adaptor
B&K Precision 1657 AC Power Source, 1500 VA AC.
More Details
Ac Power
Buy Ac Power Items Including:
1653A : 150V 2A AC Power Supply (1653A)
B&K Precision 1657 AC Power Source, 1500 VA AC
AC Power Adapter; 115 VAC
Davis Instruments AC-powered Repeater
AC Power Adapter 115VAC (PC5OPT01)
More Details
Instech Peristaltic Pump Explosion Proof
Buy Instech Peristaltic Pump Explosion Proof Items Including:
Explosion-proof Single-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/2 Hp, 3600 RPM,.
More Details
Low Rpm Ac Motor
Buy Low Rpm Ac Motor Items Including:
56C Frame AC Motor, 1725 rpm, 1/2 hp, totally enclosed fan-cooled (TEF (CL3504)
Explosion-proof Single-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM,.
More Details
Marathon Pumps
Buy Marathon Pumps Items Including:
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, (56C34F5301/ D311)
Explosion-proof Single-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM,.
More Details
1 Rpm Ac Motor
Buy 1 Rpm Ac Motor Items Including:
56C Frame AC Motor, 1725 rpm, 1/2 hp, totally enclosed fan-cooled (TEF (CL3504)
Motor, General-Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1-1/2 (G258)
Motor, General-Purpose, Single-Phase, Open Drip-Proof, AC Motor, 1-1/2 (G270).
More Details
70618
results
<<<Previous
4
5
6
7
8
9
10
11
12
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Variable Ac Power Supplies to 1 Rpm Ac Motor
>
31125329-s-toc_2 31125329stoc2