Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 800
Chlorine Tubing
to
Sensidyne Air Sampling and Gas Analysis
Image
Product Detail
More Detail
Chlorine Tubing
Buy Chlorine Tubing Items Including:
Sensidyne Gas Detection Tubes, Chlorine (SEI)
RAE Systems gas detection tubes, chlorine, 5 to 100 ppm, 10/box
RAE Systems gas detection tubes, chlorine, 0.5 to 8 ppm, 10/box
DPD Free Chlorine Reagent Powder Pillow Packs, 5 mL, 1000 Tests (RP009-3).
More Details
Drager Tubes
Buy Drager Tubes Items Including:
Drager Tubes, Acetic Acid, 5 To 80 ppm
Drager Tubes, Acetone, 10 To 12,000 ppm
Drager Tubes, Ammonia, 2.5 to 100 ppm
Drager Tubes, Carbon Dioxide, 0.1 to 6%
Drager Tubes, Carbon Dioxide, 1 to 20%
More Details
Water Sampling Tube
Buy Water Sampling Tube Items Including:
RAE Systems gas detection tubes, water vapor, 2 to 10 lbs?MMCF, 10/box
RAE Systems gas detection tubes, water vapor, 60 to 40 lbs/MMCF, 10/bo.
More Details
Hydrochloric Acid Pumps
Buy Hydrochloric Acid Pumps Items Including:
Drager Tubes, Hydrochloric Acid, 50 to 5000 ppm.
More Details
Acetic Acid 2 Ear Solution
Buy Acetic Acid 2 Ear Solution Items Including:
ACETIC ACID GLACIAL ACS 2 1/2L (YO-88051-42).
More Details
Ammonia Gas Analyzer
Buy Ammonia Gas Analyzer Items Including:
CMS Chip, Ammonia, 100 to 2000 ppm.
More Details
Polyethylene Tubing 10 Cm Diameter
Buy Polyethylene Tubing 10 Cm Diameter Items Including:
CMS Chip, Ammonia, 2 to 50 ppm
CMS Chip, Benzene, 10 to 250 ppm
CMS Chip, Vinyl Chloride, 10 to 250 ppm
Tubing,flexelene,3/8" X 1/2",100' (YO-96155-10)
Spectra/Por 4 Membranes; flat width, 10mm; diameter, 6.4mm; length, 10 (YO-02900-41).
More Details
Tubing for Acid
Buy Tubing for Acid Items Including:
Drager Tubes, Hydrochloric Acid, 1 To 10 PPm
Drager Tubes, Hydrochloric Acid, 50 to 5000 ppm
DRAGER TUBES, Hydrocyanic acid, 2 TO 150 PPM
Drager Diffusion Tube Hydrochloric Acid 10/a-D
Drager Diffusion Tube Hydrochloric Acid 20/a-D.
More Details
Benzene Pump
Buy Benzene Pump Items Including:
Drager Tubes, Benzene, 0.5 to 10 ppm.
More Details
Sampling Pumps
Buy Sampling Pumps Items Including:
Sensidyne Gas sampling pump
Global Water SP100 Fixed Speed, Peristaltic, Fluid Sampling Pump (SP100)
Gilian 3500 Air Sampling Pump Kit, 115 VAC (801934-1201)
Gilian 3500 Air Sampling Pump Kit, 230 VAC (910-0001-02)
Casella Tuff™ Air Sampling Pump.
More Details
Hydrogen Tubing
Buy Hydrogen Tubing Items Including:
Sensidyne Gas Detection Tubes, Hydrogen chloride (SEI)
Sensidyne Gas Detection Tubes, Hydrogen cyanide (SEI)
Sensidyne Gas Detection Tubes, Hydrogen sulfide (YO-81900-85)
Sensidyne Gas Detection Tubes, Hydrogen Sulfide (YO-81901-36).
More Details
Test Air Sampling and Gas Analysis
Buy Test Air Sampling and Gas Analysis Items Including:
Combustible Gas Leak Detector with Alarm
Sensidyne Gas Detection Tubes, Benzene (SEI, Twin Tubes (5 Tests/Box))
Sensidyne Gas Detection Tubes, Nitrogen oxide (Twin Tubes (5 Tests/Box
Sensidyne Gas Detection Tubes, Oxygen (Twin Tubes (5 Tests/Box)).
More Details
Gas Detection Systems
Buy Gas Detection Systems Items Including:
RAE Systems gas detection tubes, carbon dioxide, 300 to 5000 ppm, 10/b
RAE Systems gas detection tubes, hydrogen chloride, 1 to 20 ppm, 10/bo
RAE Systems gas detection tubes, hydrogen fluoride, 0.5 to 20 ppm, 10/.
More Details
Drager Gas Detection-Tubes
Buy Drager Gas Detection-Tubes Items Including:
Drager Tubes, Acetic Acid, 5 To 80 ppm
Drager Tubes, Acetone, 10 To 12,000 ppm
Drager Tubes, Hydrogen Sulfide, 2 To 200 PPm
Drager Tubes, Hydrogen Sulfide, 0 to 200 ppm
Drager Tubes, Nitrogen Dioxide, 0.5 to 25 ppm
More Details
5 Hydrochloric Acid Solution for Cleaning
Buy 5 Hydrochloric Acid Solution for Cleaning Items Including:
Drager Tubes, Hydrochloric Acid, 1 To 10 PPm
Drager Tubes, Hydrochloric Acid, 50 to 5000 ppm
Drager Diffusion Tube Hydrochloric Acid 10/a-D
Drager Diffusion Tube Hydrochloric Acid 20/a-D
5-NITROSALICYLIC ACID 99% 250G (YO-88276-51).
More Details
Aim of Ethylene Oxide Gas
Buy Aim of Ethylene Oxide Gas Items Including:
Sensidyne Gas Detection Tubes, Ethylene oxide
Drager Tubes, Ethylene Oxide, 1 To 15 ppm
Portable Ethylene Oxide Detector, 0 to 10 ppm
CMS Chip, Ethylene Oxide, 0.4 to 5.0 ppm
Long sterilizer system, 3.0 cu ft 115 VAC, 60 Hz (AN74IX STARTER KIT).
More Details
Gas Test
Buy Gas Test Items Including:
Combustible Gas Leak Detector with Alarm
Sensidyne Gas Detection Tubes, Benzene (SEI, Twin Tubes (5 Tests/Box))
Sensidyne Gas Detection Tubes, Nitrogen oxide (Twin Tubes (5 Tests/Box
Sensidyne Gas Detection Tubes, Oxygen (Twin Tubes (5 Tests/Box)).
More Details
Fluoride Ppm
Buy Fluoride Ppm Items Including:
RAE Systems gas detection tubes, hydrogen fluoride, 0.5 to 20 ppm, 10/
DRAGER TUBES, Hydrogen Fluoride, 0.5 TO 90 PPM
Fluoride Standard 1000 PPM.
More Details
Hydrogen Gas Detection
Buy Hydrogen Gas Detection Items Including:
Sensidyne Gas Detection Tubes, Hydrogen chloride (SEI)
Sensidyne Gas Detection Tubes, Hydrogen cyanide (SEI)
Drager Tubes, Hydrogen Sulfide, 2 To 200 PPm
Drager Tubes, Hydrogen Sulfide, 0 to 200 ppm
DRAGER TUBES, Hydrogen Sulfide, 100 TO 2000 PPM.
More Details
Chlorine Gas Detection
Buy Chlorine Gas Detection Items Including:
Sensidyne Gas Detection Tubes, Chlorine
RAE Systems gas detection tubes, chlorine, 5 to 100 ppm, 10/box
RAE Systems gas detection tubes, chlorine, 0.5 to 8 ppm, 10/box
Drager Tubes, Chlorine, 0.2 to 30 ppm
Portable Chlorine Detector, 0 to 20 ppm.
More Details
Acid Tubing Pump Laboratory
Buy Acid Tubing Pump Laboratory Items Including:
Drager Tubes, Acetic Acid, 5 To 80 ppm
Drager Tubes, Hydrochloric Acid, 1 To 10 PPm
Drager Tubes, Hydrochloric Acid, 50 to 5000 ppm.
More Details
Polyethylene Tubing Water Vapor Permeability
Buy Polyethylene Tubing Water Vapor Permeability Items Including:
RAE Systems gas detection tubes, water vapor, 2 to 10 lbs?MMCF, 10/box.
More Details
Ethylene Oxide Gas Supplier California
Buy Ethylene Oxide Gas Supplier California Items Including:
Sensidyne Gas Detection Tubes, Ethylene oxide
Drager Tubes, Ethylene Oxide, 1 To 15 ppm
Portable Ethylene Oxide Detector, 0 to 10 ppm
CMS Chip, Ethylene Oxide, 0.4 to 5.0 ppm
Long sterilizer system, 3.0 cu ft 115 VAC, 60 Hz (AN74IX STARTER KIT).
More Details
Ethylene Gas
Buy Ethylene Gas Items Including:
Sensidyne Gas Detection Tubes, Ethylene oxide
Sensidyne Gas Detection Tubes, Ethylene (YO-81900-82)
Sensidyne Gas Detection Tubes, Ethylene (YO-81901-30)
Drager Tubes, Ethylene Glycol, 10 to 180 mg/m3
Portable Ethylene Oxide Detector, 0 to 10 ppm.
More Details
Rae Systems Gas Detection Tubes
Buy Rae Systems Gas Detection Tubes Items Including:
RAE Systems gas detection tubes, acetone, 0.1 to 2%, 10/box
RAE Systems gas detection tubes, ammonia, 5 to 100 ppm, 10/box
RAE Systems gas detection tubes, ammonia, 1 to 15%, 10/box
RAE Systems gas detection tubes, carbon dioxide, 1 to 20%, 10/box.
More Details
Fluoride Analysis
Buy Fluoride Analysis Items Including:
Sensidyne Gas Detection Tubes, Hydrogen fluoride
DRAGER TUBES, Hydrogen Fluoride, 0.5 TO 90 PPM
ISE electrode, fluoride, for controller 56105-00 (YO-27077-06)
Kit repl solid state for fluoride electrode (YO-27506-80)
Fluoride Standard 1000 PPM.
More Details
Hydrocarbon Detection
Buy Hydrocarbon Detection Items Including:
Sensidyne Gas Detection Tubes, Hydrocarbon, high-class
RAE Systems gas detection tubes, hydrocarbons, 50 to 1000 ppm, 10/box
DRAGER TUBES, Petroleum Hydrocarbons, 100 TO 2500 PPM
Portable Hydrocarbon (non-methane) Detector, 0 to 100% LEL.
More Details
Petroleum Sampling
Buy Petroleum Sampling Items Including:
DRAGER TUBES, Petroleum Hydrocarbons, 10 TO 300 PPM
DRAGER TUBES, Petroleum Hydrocarbons, 100 TO 2500 PPM
Sample Vial Holder for Carbon Residue Tester, six places for 16-mL via
Sample Vial Holder for Carbon Residue Tester, seven places for 2-mL an.
More Details
Chloroform To
Buy Chloroform To Items Including:
BENZYL CHLOROFORMATE 97 WT 5G (YO-88155-91)
CHLOROFORM-D 99.8 ATOM % 10ML (YO-88232-26)
CHLOROFORM-D 100 ATOM % D 5ML (YO-88232-27)
CHLOROFORM-D 100 ATOM % 7.5ML (YO-88291-64)
CHLOROFORM-D 99.8 ATOM % 7.5ML (YO-88291-65).
More Details
Sensidyne Air Sampling and Gas Analysis
Buy Sensidyne Air Sampling and Gas Analysis Items Including:
Sensidyne Gas sampling pump
Sensidyne Gas Detection Tubes, Mercury vapor
Sensidyne Gas Detection Tubes, Nitrogen oxide (Twin Tubes (5 Tests/Box
Sensidyne Gas Detection Tubes, Oxygen (Twin Tubes (5 Tests/Box))
Sensidyne Gas Detection Tubes, Xylene.
More Details
70618
results
<<<Previous
796
797
798
799
800
801
802
803
804
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
700
710
720
730
740
750
760
770
780
785
786
787
788
789
790
791
792
793
794
795
796
797
798
799
800
801
802
803
804
805
806
807
808
809
810
811
812
813
814
820
830
840
850
860
870
880
890
900
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
2,000
2,354
Chlorine Tubing to Sensidyne Air Sampling and Gas Analysis
>
31125329-s-toc_2 31125329stoc2