Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 1
18 High Temperature Gloves
to
Water Bath Chiller
Image
Product Detail
More Detail
18 High Temperature Gloves
Buy 18 High Temperature Gloves Items Including:
Zetex High-Temperature Gloves, up to 1100°F, one size fits all, 14
ZetexPlus High-Temperature Gloves, up to 2000°F, one size fits all
Zetex High-Temperature Finger Cots, up to 1100°F, one size fits al
Whirl-Pak 18 oz High Temperature Bags, 500/box.
More Details
Neslab Chiller
Buy Neslab Chiller Items Including:
Thermo Scientific NESLAB ThermoFlex™ 900 Recirculating Chiller with (YO-13042-00)
Thermo Scientific NESLAB ThermoFlex™ 2500 IPR Recirculating Chiller (YO-13042-42)
Thermo Scientific NESLAB ThermoFlex™ 2500 IPR Recirculating Chiller (YO-13042-44).
More Details
Genesys 10 Uv Scanning Spectrophotometer
Buy Genesys 10 Uv Scanning Spectrophotometer Items Including:
GENESYS 10 nanoCell accessory, 0.2mm pathlength adapter (222-227900)
GENESYS 10 nanoCell accessory, 1.0mm pathlength adapter (222-228000)
GENESYS 10 nanoCell accessory with 0.2mm and 1.0mm pathlength adapter (222-230300).
More Details
Low Temperature Tube Furnace
Buy Low Temperature Tube Furnace Items Including:
Horizontal Tube Furnace, 18-1/2" Tube Length, 1600°C, 208 / 240 V (RHTH 120/150/16/C42)
Horizontal Tube Furnace, 18-1/2" Tube Length, 1700°C, 208 / 240 V (RHTH 120/150/17/C42).
More Details
Thermo Scientific Fume Hoods
Buy Thermo Scientific Fume Hoods Items Including:
Standing Height Wall Assembly with Fume Hood - 13ft (YO-41524-50)
Complete 4foot Fume Hood System With Acid Storage Cabinet Epoxy Worksu (YO-41525-00)
Complete 4foot Fume Hood System With General Base Storage Cabinet Epox (YO-41525-05).
More Details
Temperature and Humidity Chambers
Buy Temperature and Humidity Chambers Items Including:
Temperature And Humidity Chamber 20 L, (0 C To 120°C) 20L; 220V 60
Temperature And Humidity Chamber 65 L, (0 C To 120°C) 65L; 220V 60
Temperature And Humidity Chamber 20 L, (0 C To 120°C) 20L; 230V 50.
More Details
Cowles Blade Lab Mixers
Buy Cowles Blade Lab Mixers Items Including:
Compact straight blade impeller; 1-1/4" dia
Sanitary Mixer, 1/2" Tri-clamp size, 6 elements (YO-04668-28)
Sanitary Mixer, 3/4" Tri-clamp size, 6 elements (YO-04668-32)
Sanitary Mixer, 1" Tri-clamp size, 6 elements (YO-04668-36).
More Details
Water Deionizer
Buy Water Deionizer Items Including:
Thermo Scientific Barnstead E-pure™ 3-Holder Water Purification Sys (D4632-33)
Thermo Scientific Bantam Deionizer, 120V (YO-99261-01)
Monobed deionizer, large capacity; 110 VAC (AD-1400)
Monobed deionizer, large-capacity; 220 VAC (AD-1400 220VAC).
More Details
Thermo Neslab Chiller
Buy Thermo Neslab Chiller Items Including:
Thermo Scientific NESLAB ThermoFlex™ 900 Recirculating Chiller with (YO-13042-00).
More Details
Thermo Scientific Revco Elite Plus Freezer
Buy Thermo Scientific Revco Elite Plus Freezer Items Including:
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-01)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-02)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03).
More Details
Oakton Ph Buffer Coa
Buy Oakton Ph Buffer Coa Items Including:
Oakton pH buffer pouches; pH 4.01, 20/pk
Oakton pH buffer pouches; pH 7.00, 20/pk
Oakton pH buffer pouches; pH 10.00, 20/pk
Oakton 4.01 pH buffer, 500-mL
Oakton 1.68 pH buffer, 500-mL
More Details
Large Peristaltic Pump
Buy Large Peristaltic Pump Items Including:
Pump, Cole-Parmer Large Peristaltic Metering Pump, Max flow of 54 GPH, (YO-74203-64)
Pump, Cole-Parmer Large Peristaltic Metering Pump, Max flow of 100 GPH (YO-74203-66).
More Details
Laboratory Refrigerators
Buy Laboratory Refrigerators Items Including:
Nor-Lake Scientific Select™ Pass-thru Refrigerator +4C, Four door 2
Nor-Lake Scientific Select™ Pass-thru Refrigerator +4C, Two Glass D
Laboratory undercounter refrigerator (single door); 5.4 CU
FT; 115V (REL-404A).
More Details
Vacuum Regulator
Buy Vacuum Regulator Items Including:
Vacuum regulator, 0 to 29" Hg, 2.47cfm (AI3600003)
Vacuum Regulator, 0 to 30" Hg, 2 cfm Max Capacity (AJ675)
Pressure Regulator, 0 to 125 psig, 20 cfm Capacity (R262)
Pressure Regulator, 0 to 100 psig, 2 cfm Capacity (AJ674).
More Details
Benchtop Fume Hood
Buy Benchtop Fume Hood Items Including:
Ductless, benchtop fume hood; 115 VAC/60 Hz (YO-33730-10)
Complete 4foot Fume Hood System With Acid Storage Cabinet Epoxy Worksu (YO-41525-00)
Complete 4foot Fume Hood System With General Base Storage Cabinet Epox (YO-41525-05).
More Details
Muffle Furnace Thermo Scientific
Buy Muffle Furnace Thermo Scientific Items Including:
Thermo Scientific Thermolyne Atmosphere-Controlled Programmable Muffle (F6028C-60-80)
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in ch (F47915)
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in ch (F47910-33).
More Details
Fisher Ph Meter Field
Buy Fisher Ph Meter Field Items Including:
Accumet XL150 pH Benchtop Meter Only
Accumet XL150 pH Benchtop Meter Kit
accumet AB150 pH Benchtop Meter Only
Accumet AB200 pH/Conductivity Benchtop Meter Only
Accumet AB150B pH Benchtop Meter Kit
More Details
Recirculating Water Chiller
Buy Recirculating Water Chiller Items Including:
Cole-ParmerRefrigerated Chillers with Immersion Probe; range, -60 to 2 (YO-01283-60)
Thermo Scientific NESLAB ThermoFlex™ 900 Recirculating Chiller with (YO-13042-00)
Thermo Scientific NESLAB ThermoFlex™ 900 Recirculating Chiller with (YO-13042-06).
More Details
Ph Conductivity Meter
Buy Ph Conductivity Meter Items Including:
Thermo Scientific Orion 4-Star pH/Conductivity Meter Only
Thermo Scientific Orion 4-Star pH/Conductivity Meter Kit
accumet® XL20 pH/Conductivity Meter
Hach ISFET Benchtop pH/Conductivity Meter Only
Hach Waterproof ISFET pH/Conductivity Meter Only.
More Details
Micro 90 Detergent
Buy Micro 90 Detergent Items Including:
Cole-Parmer® Micro-90® cleaning solution, case of twelve 1 qt (YO-18100-00)
Cole-Parmer Micro-90 cleaning solution, 55 gallon drum
Cole-Parmer Micro-90 cleaning solution, 1 liter bottle
Detergent 8 Cleaning Detergent, case of four 1-gallon bottles.
More Details
150 Psi Micro Water Pump
Buy 150 Psi Micro Water Pump Items Including:
Mini Lubricator with bowl guard, Max 32 SCFM, Max Pressure of 150 PSI,
Compact Air Filter, 5 micron filter, Max 73 SCFM, Max Pressure 150 PSI
Thermo Scientific Nalgene Aspirator pump, 7.5 psi minimum water pressu.
More Details
Revco Refrigerator
Buy Revco Refrigerator Items Including:
Laboratory undercounter refrigerator (single door); 5.4 CU
FT; 115V (REL-404A)
Laboratory space-saver refrigerator (single door); 11.5 CU FT; 115V (REL-1204A-20)
Laboratory space-saver refrigerator (single door); 11.5 CU FT; 200/230 (REL-1204D).
More Details
Large Capacity Lab Glassware Washer
Buy Large Capacity Lab Glassware Washer Items Including:
Undercounter Glassware Washer 115V (YO-98950-01)
Freestanding Glassware Washer, 115V (YO-98950-11)
Large Capacity Stainless Steel Ultrasonic Cleaners, 10 gal, 230VAC (CPN-908-013)
FlaskScrubber Undercounter Glassware Washer, 115V 60HZ (4420320).
More Details
Benchtop Autoclaves
Buy Benchtop Autoclaves Items Including:
Autoclavable Polypropylene Bags, 10 X 16 250/pk (YO-06501-02)
Autoclavable Polypropylene Bags, 16 X 18 250/pk (YO-06501-04)
Autoclavable Polypropylene Bags, 12 X 24 250/pk (YO-06501-06)
Autoclavable Polypropylene Bags, 22 X 30 250/pk (YO-06501-08).
More Details
6 Cu Ft Refrigerator
Buy 6 Cu Ft Refrigerator Items Including:
ADA Compliant General Purpose Undercounter Refrigerator, 6 cu ft (6CAD)
Flammable Material Undercounter Refrigerator, 6 cu ft (6FAR)
Hazardous Material Undercounter Refrigerator, 6 cu ft (6EAR)
General Purpose Undercounter Freezer, 6 cu ft (6CAF).
More Details
Explosion Proof Fume Hood
Buy Explosion Proof Fume Hood Items Including:
Explosion-proof air blower, 1/6 hp, 115 VAC/60 Hz
Explosion-proof air blower, 1/4 hp, 115 VAC/60 Hz (YO-09078-60)
Explosion-proof air blower, 1/4 hp, 115 VAC/60 Hz (YO-09078-90)
48" Explosion-Proof Bypass Fume Hood, 115V/60Hz (YO-09012-16).
More Details
High Temperature Lab Oven
Buy High Temperature Lab Oven Items Including:
Electronic-controlled high-temperature vacuum oven, 0.67 cu ft, 120 VA (YO-05017-00)
Microprocessor-controlled high-temperature vacuum oven, 1.50 cu ft, 12 (YO-05017-10)
High-temperature industrial mechanical convection oven, 4.3 cu ft, 115.
More Details
Lab Chillers
Buy Lab Chillers Items Including:
Cole-Parmer Flow-through Refrgerated Chillers; range, -25 to 40°C; (YO-01283-40)
Cole-Parmer Flow-through Refrgerated Chillers; range, -25 to 40°C; (YO-01283-41)
Thermo Scientific NESLAB ThermoFlex™ 5000 IPR Recirculating Chiller (YO-13042-60).
More Details
Multi Channel Peristaltic Pump
Buy Multi Channel Peristaltic Pump Items Including:
L/S 4-channel multichannel pump head for microbore tubing (YO-07534-04)
L/S 8-channel multichannel pump head for microbore tubing (YO-07534-08)
L/S 4-channel multichannel pump head for L/S tubing
L/S 8-channel multichannel pump head for L/S tubing.
More Details
Water Bath Chiller
Buy Water Bath Chiller Items Including:
Thermo Scientific NESLAB ThermoFlex™ 1400 EPR Recirculating Chiller (YO-13042-28)
Thermo Scientific NESLAB ThermoFlex™ 1400 EPR Recirculating Chiller (YO-13042-30)
Thermo Scientific NESLAB ThermoFlex™ 1400 EPR Recirculating Chiller (YO-13042-32).
More Details
70618
results
1
2
3
4
5
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
18 High Temperature Gloves to Water Bath Chiller
>
31125329-s-toc_2 31125329stoc2