Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 10

Ph Meter with Analog Output   to   Tip Sonicator

Image
  Product Detail
More Detail

Ph Meter with Analog Output

Buy Ph Meter with Analog Output Items Including:
  • Extech Light Meter with Analog Output
  • Vaisala DEWPOINT METER with Probe, Software, Analog Output Cable, AC (DM70D5B1A1B)
  • Cole-Parmer laboratory pH/mV meter, analog
  • Cole-Parmer field pH/mV meter, analog
  • MYRON L EP11/PH Analog pH/Conductivity Meter.

Hydrogen Rated Gas Solenoid Valve

Buy Hydrogen Rated Gas Solenoid Valve Items Including:
  • Solenoid Valve, 316 SS, with 1 1/4 NPT(F) Port
  • Variable-speed gas piston pump, Class 1, Division 2 rated for hazardou
  • Variable-speed gas piston pump, Class 1, Division 1 rated for hazardou
  • Variable-speed rate controller, standard, 115 VAC.

Venturi Vacuum Pump Water Powered

Buy Venturi Vacuum Pump Water Powered Items Including:
  • Miniature venturi vacuum pump, 0.5 cfm, 28.0
  • Filter Pack For 35031-00 & -10 Vacuum Pumps, 5 Pack (72200)
  • VENTURI with AIR SAVER technology,28" Hg,1.2 CFM, With Gauge
  • Global Water SP100 Fixed Speed, Peristaltic, Fluid Sampling Pump (SP100).

Sanplatec Dry Keeper Desiccator

Buy Sanplatec Dry Keeper Desiccator Items Including:
  • Dry-Keeper™ Desiccator Cabinets, Horizontal; Non-Electric (H420580001)
  • Dry-Keeper™ Desiccator Cabinets, Vertical; Non-Electric (H420560001)
  • Dry-Keeper™ Desiccator Cabinets, Horizontal; Electronic (H420580003).

Headspace Oxygen Analyzer

Buy Headspace Oxygen Analyzer Items Including:
  • Handheld Headspace Oxygen Analyzer
  • Benchtop Headspace Oxygen Analyzer, 100-240VAC
  • Replacement Sampling Probe Assembly (9007)
  • Replacement Needles, Side Port Hole; 3/pack (9003)
  • Replacement Septa; 200/pack (9001).

Nist Certified Thermometer Fisher Scientific

Buy Nist Certified Thermometer Fisher Scientific Items Including:
  • Easy-read NIST Certified Thermometer, -20/110°C, 300mm Length, Tot (30500C)
  • Easy-read NIST Certified Thermometer, -10/225c, 355mm Length, Total Im (30520C)
  • Durac Plus Thermometer, Certified To NIST, -20/110°C, 300mm Length (3/4110).

Hand Vacuum Pump Water Sampling

Buy Hand Vacuum Pump Water Sampling Items Including:
  • Hand-operated vacuum pump, 16 mL/stroke, PVC body
  • Hand-operated vacuum pump (without gauge), 32 mL/stroke, PVC body
  • Global Water SP100 Fixed Speed, Peristaltic, Fluid Sampling Pump (SP100)
  • Global Water WS700 Peristaltic Water Sampler (YO-99243-70).

Orion Ph Meters

Buy Orion Ph Meters Items Including:
  • Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9102BNWP)
  • Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9103BNWP)
  • Thermo Scientific Orion pH electrode, glass body, refillable, single-j (9104BNWP).

Aluminum Weighing Dishes

Buy Aluminum Weighing Dishes Items Including:
  • Aluminum Weighing Dishes, 73mm Dia x 11/16" D, 2.5 gr., 80ml Capacity, (YO-01018-29)
  • Aluminum Weighing Dishes, 103mm Dia x 3/4" D, 7.0 gr., 150ml Capacity, (YO-01018-31)
  • Micro Aluminum Weighing Dishes, 6mm Dia
  • x 2.5mm H, .035ml, 250/pk.

Multi Channel Thermocouple Meter

Buy Multi Channel Thermocouple Meter Items Including:
  • QuadTemp2000: Four-Channel Thermocouple Recorder with LCD (QuadTemp2000)
  • OctTemp2000: Four-Channel Thermocouple Recorder with LCD (OctTemp2000)
  • YSI MultiLab Three-Channel Multi-parameter Benchtop Meter
  • 4-Channel Thermometer w/ SD Card Data Logging.

Orion Ph Electrode Storage Solution 910001

Buy Orion Ph Electrode Storage Solution 910001 Items Including:
  • Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)
  • Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottl
  • Oakton electrode storage solution, case of 24
  • Thermo Scientific Orion ROSS Storage Soluton, 475-mL bottle.

Aspirator Pump

Buy Aspirator Pump Items Including:
  • Stainles Steel Aspirator for 35031-00 & -10, 1 Each (71121)
  • Cooling Coil for 35031-00 and -10 vacuum pumps (71501)
  • Filter Pack For 35031-00 & -10 Vacuum Pumps, 5 Pack (72200)
  • Filter Pack For 35031-00 & -10 Vacuum Pumps, 10 Pack (72210).

Iq Scientific Instruments Ph Meter

Buy Iq Scientific Instruments Ph Meter Items Including:
  • Thermo Scientific Orion 3-Star handheld pH meter kit
  • Eutech Instruments pH 2000 1/2-DIN Wall- Mount pH/ORP Controller
  • Eutech Instruments pH 2000 1/4-DIN Panel-Mount pH/ORP Controller
  • Eutech Instruments pH 500 2-Wire pH/ORP Transmitter with Display.

Turbidity Standards

Buy Turbidity Standards Items Including:
  • Turbidity Standard, 1.0 NTU, 1-L bottle
  • Turbidity Standard, 10.0 NTU, 1-L bottle
  • Turbidity Standard, 40.0 NTU, 1-L bottle
  • Turbidity Standard, 0.2 NTU, 1-L bottle
  • Turbidity Standards Kit for Cole-Parmer and ICM meters, 0.5, 5.0, 40 N (YO-08391-14).

Oxford Benchmate Adjustable Volume Pipette

Buy Oxford Benchmate Adjustable Volume Pipette Items Including:
  • Ovation Adjustable-Volume Pipette, 0.2 to 2.0 µL, Red
  • Ovation Adjustable-Volume Pipette, 1 to 10 µL, Orange
  • Ovation Bionatural Adjustable Volume Pipette, Left-handed, 100-5000mu
  • Ovation Bionatural Adjustable Volume Pipette, Left-handed, 0.2 - 10mL.

Salinity and Ph Meter

Buy Salinity and Ph Meter Items Including:
  • Oakton SALT 6+ handheld salinity meter with probe and NIST-traceable C
  • Oakton SALT 6+ handheld salinity meter kit with NIST-traceable Calibra
  • Cole-Parmer® Salinity Pocket Tester
  • Thermo Scientific Orion Star A215 pH/Conductivity Benchtop Meter Only.

Vacuum Gauge Torr

Buy Vacuum Gauge Torr Items Including:
  • 0.01 to 20 torr Cole-Parmer Pressure/Vacuum Meter for Pirani-Type Sens
  • 1 to 1500 torr Cole-Parmer Pressure/Vacuum Meter for Diaphragm-Type Se
  • 0.01 to 20 torr Cole-Parmer Pressure/Vacuum Controller for Pirani-Type.

Laboratory Stands

Buy Laboratory Stands Items Including:
  • Bag Opener Stand For Holding Bags Size 400 mL, 1 Each (211-040)
  • Free Standing Spindle-Ready Washer, 230V
  • Support Stand Stainless steel with 28” Rod
  • Polypropylene Coated Metal Support Stand with Center Rod
  • Heavy Duty support stand with 23” s/s rod.

Eutech Ph Meter

Buy Eutech Ph Meter Items Including:
  • Eutech pH 5+ pH meter kit (YO-15940-79)
  • Eutech pH 6+ pH/ORP meter kit (YO-15940-80)
  • Power Adapter 110/240V for 1500 pH Meter (YO-99855-90)
  • Eutech Instruments 1/4-din pH 800 On/off Controller, 115 Vac
  • Eutech Instruments pH 1000 controller, 115 VAC.

Air or Water Driven Magnetic Stirrer

Buy Air or Water Driven Magnetic Stirrer Items Including:
  • Bath for Magnetic Stirrer, 3.8L, 120VAC
  • Bath for Magnetic Stirrer, 3.8L, 220VAC
  • Bath for Magnetic Stirrer, 8L, 120VAC
  • Bath for Magnetic Stirrer, 12L, 120VAC
  • Bath for Magnetic Stirrer, 12L, 220VAC

100 Micron Nylon Filter Cloth

Buy 100 Micron Nylon Filter Cloth Items Including:
  • Nylon monofilament filter bag; 10"L, 100 µm (255009-03)
  • Nylon monofilament filter bag; 18-1/2"L, 100 µm (255032-03)
  • Spectra/Mesh Screen Discs, 90 mm Nylon, 90mm dia; pore size, 100µ;
  • Spectra/Mesh Screen Discs, 55 mm, Nylon, 100 µm, 10/pk.

Sorvall Centrifuge Tubes

Buy Sorvall Centrifuge Tubes Items Including:
  • Adapters For 100ml Round Bottom Open-top Tubes (2 Positions), Fits Buc (75003713)
  • Adapters For 50ml Conical Tubes (3 Positions) And Sealing Vessel, Fits (75003714)
  • Adapters For 50ml Conical Tubes (7 Positions), Fits Bucket 17708-01, 4 (75003638).

Honeywell Temperature Controller

Buy Honeywell Temperature Controller Items Including:
  • Temperature Controller, Unversal Input, 1/4-DIN, Relay Output, Alarm R (DC2500EB0A0020000000000)
  • Temperature Controller, Unversal Input, 1/4-DIN, Current Output, Alarm (DC2500CB0A0020000000000).

Vacuum Chart Recorders

Buy Vacuum Chart Recorders Items Including:
  • Absolute Pressure Module 14.5 psi
  • Absolute Pressure Module 43.5 psi
  • Absolute Pressure Module 1450 psi
  • Absolute Pressure Module 4350 psi
  • Absolute Pressure Module 14,500 psi

Small Electric Fluid Transfer Pumps

Buy Small Electric Fluid Transfer Pumps Items Including:
  • 6/10 Rack for Cole-Parmer Syringe Pump, Infusion and Withdrawal design (788300) (YO-74903-60)
  • 4 Multi Rack for Cole-Parmer Syringe Pump, Infusion and Withdrawal des (788301) (YO-74903-61).

Water Bath Stirrer

Buy Water Bath Stirrer Items Including:
  • Standard magnetic stirrer, 50mL to 3L; 120V, 60Hz
  • Standard magnetic stirrer, 100mL to 7L (YO-02918-14)
  • 6-position magnetic stirrer, 30mL to 2L (YO-02918-18)
  • Standard magnetic stirrer, 50mL to 3L; 230V, 50Hz
  • Bath for Magnetic Stirrer, 3.8L, 120VAC.

Rosemount Cold Water Ph Probe

Buy Rosemount Cold Water Ph Probe Items Including:
  • Oakton Waterproof pH 600 Meter with pH probe
  • Oakton Waterproof pH 610 Meter with pH probe with NIST-traceable Calib
  • Oakton Waterproof pH 620 Meter with pH probe with NIST-traceable Calib
  • Waterproof ISFET pH probe, heavyduty piercing tip (PHW57-SS).

Nalgene Narrow Mouth Bottle Hdpe

Buy Nalgene Narrow Mouth Bottle Hdpe Items Including:
  • Thermo Scientific Nalgene narrow-mouth bottle of PTFE ® FEP, 64oz 2/p
  • Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, HDPE, 30 mL, 12 (YO-06035-07)
  • Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, HDPE, 30 mL, 72 (YO-06035-09).

Heavy Metal Water Testing Meter

Buy Heavy Metal Water Testing Meter Items Including:
  • Heavy-duty water softening system with meter regeneration; 6 GPM, 18,6 (162405 (W-422F))
  • Heavy-duty water softening system with meter regeneration; 9 GPM, 24,8 (162406 (W-430F))
  • Watersafe Well Water Test Kit
  • LaMotte Ammonia Test Strips, 0 to 6 ppm.

Tip Sonicator

Buy Tip Sonicator Items Including:
  • Probe, 1/2", with replaceable tip
  • Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1/2"
  • Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1" di
  • Ultrasonic Cell Disruptor Replacement Tips, 1/2" dia.



Ph Meter with Analog Output to Tip Sonicator

>