Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 11

Liquid Sampling   to   5000 Ml Separatory Funnel

Image
  Product Detail
More Detail

Liquid Sampling

Buy Liquid Sampling Items Including:
  • Permanently Sealed Dispersive Cells for Liquid Samples; window materia (5153)
  • Permanently Sealed Dispersive Cells for Liquid Samples; window materia (5156)
  • Drum and Liquid Sampler, 3', HDPE (114505)
  • Manning Peristaltic Sampling Pump (PST8B2A1A4B2).

Oakton Ph 110

Buy Oakton Ph 110 Items Including:
  • Oakton advanced pH 110 meter only
  • Oakton advanced pH 110 meter only with NIST-traceable Calibration
  • Oakton advanced pH 110 meter kit with NIST-traceable Calibration
  • Oakton advanced pH 110 meter with probe
  • Oakton DO 110 Dissolved Oxygen Meter Only.

Laboratory Stand Mixer

Buy Laboratory Stand Mixer Items Including:
  • Bag Opener Stand For Holding Bags Size 80 And 100 mL, 1 Each (211-010)
  • Mixer plate stand, 22"
  • Mixer plate stand, 31 1/2"
  • Mixer H-stand, adjustable to 39 3/4"
  • Complete Mixer System, 45-2000RPM 115V with stand and clamp (YO-54401-05).

Probes for Q55 Sonicator Ultrasonic Processor

Buy Probes for Q55 Sonicator Ultrasonic Processor Items Including:
  • Microprobe, 1/4", 10 - 50ml, for use with Q55 Sonicator
  • Coupler (for Probe 04712-65) for use with ultrasonic processors
  • Dual Horn (with two 3/4" solid probes) for use with ultrasonic process
  • Probe, 1", solid
  • Probe, 1/2", solid.

Accumet Ph Meters and Probes

Buy Accumet Ph Meters and Probes Items Including:
  • Accumet Benchtop Meter Replacement pH/ATC Probe
  • Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
  • Accumet pH Electrode, Combination, General Purpose, Long-neck, Gel-fil
  • Oakton pH 700 Benchtop Meter, meter and probe stand.

Laboratory Sample Storage Refrigerator

Buy Laboratory Sample Storage Refrigerator Items Including:
  • Kimble Standard SSr™ Vial, 2mL, 1000/cs (60680-12)
  • Kimble Straight-sided SSr™ Vial, 2ml, 1000/cs (60690-12)
  • Kimble Straight-sided SSr™ Vial, 4ml, 1000/cs (60690-1)
  • Kimble Straight-sided SSr™ Vial, 7mL, 1000/cs (60690-2).

Qsonica Accessories for Sonicator Ultrasonic Processors

Buy Qsonica Accessories for Sonicator Ultrasonic Processors Items Including:
  • Open end wrench (requires 2) for use with ultrasonic processors
  • Replacement tube holder (20 tubes) for use with ultrasonic processors
  • Dual Horn (without probes) for use with ultrasonic processors
  • Stand with Clamp for use with ultrasonic processors.

Plastic Bulkhead Fittings 2

Buy Plastic Bulkhead Fittings 2 Items Including:
  • Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/2" tubing ID
  • Bulkhead Fitting, PVDF, 1/2" NPT(F) Port X 2-1/16" OD (FK-TA04)
  • Bulkhead Fitting, PVDF, 1-1/4" NPT(F) Port X 2-7/8" OD (FK-TA10).

Peristaltic Tubing Types

Buy Peristaltic Tubing Types Items Including:
  • Replacement Tubing Set for Cole-Parmer 73160-XX pumps, Silicon, 0.8, 1
  • Replacement Tubing Set for Cole-Parmer 73160-XX pumps, Viton, 0.8, 1.6
  • Masterflex C/L Dual-Channel Variable-Speed Tubing Pump; 10 to 60 rpm, (YO-77120-42).

Mechanical Force Gauges

Buy Mechanical Force Gauges Items Including:
  • FB-100 : Force Gauge, 100LBF Capacity Mechanical Force Gauge
  • Mechanical force gauge, 2 lb capacity (MF-2)
  • Mechanical force gauge, 1 kg capacity (MF-1KG)
  • Mechanical force gauge, 2 kg capacity (MF-2KG)
  • Mechanical force gauge, 10 lb capacity (MF-10).

12 Oz Plastic Bottles

Buy 12 Oz Plastic Bottles Items Including:
  • Reaction vessel with coupling, 12 oz
  • Multi-port stirring assembly with 12-oz reaction vessel
  • Wide-Mouth Sample Containers, PP, 1.2 L
  • Mark-10 MR53-12 Torque Sensor, 12 lb x 192 oz in x 135 N cm
  • Cole-Parmer Bottle, Clear Boston Round, 2 oz, 24/case.

Cga 350 Regulator

Buy Cga 350 Regulator Items Including:
  • Single-Stage Regulator, 2000 scfh Capacity, 350 CGA Fitting (YO-98202-02)
  • Two-Stage High-Purity Gas Regulator, 270 scfh Capacity, 350 CGA (YO-98210-02)
  • Two-Stage High-Purity Gas Regulator, 380 scfh Capacity, 350 CGA (YO-98210-12).

Pyrex Test Tubes

Buy Pyrex Test Tubes Items Including:
  • Pyrex Brand 9800 test tube; 14 mL, pack of 72
  • Pyrex Brand 9800 test tube; 20 mL, pack of 72
  • Pyrex Brand 9800 test tube; 27 mL, pack of 72
  • Pyrex Brand 9800 test tube; 34 mL, pack of 72
  • Pyrex Brand 9800 test tube; 55 mL, pack of 72.

Thermo Scientific Nalgene Labware

Buy Thermo Scientific Nalgene Labware Items Including:
  • Thermo Scientific Nalgene high-density polyethylene narrow-mouth sampl (2089-0001)
  • Thermo Scientific Nalgene high-density polyethylene narrow-mouth sampl (2089-0002)
  • Thermo Scientific Nalgene high-density polyethylene narrow-mouth sampl (2089-0004).

1 Liter Hdpe Plastic Bottles

Buy 1 Liter Hdpe Plastic Bottles Items Including:
  • Cole-Parmer Bottletop Dispenser 1-10 mL with 250 mL Plastic Bottle
  • Qorpak Kaptclean™ pre-cleaned sample bottle, 1 liter
  • Stainless steel sanitary bottle; 1 liter, 2" flange (PS-10F)
  • Nutrient broth, prepared; 1 liter bottle, 10/pk (YO-14204-74).

Microfluidic Products

Buy Microfluidic Products Items Including:
  • Microfluidic Slide Connector Set, includes two type 4x2, four type 2x2
  • Microfluidic Micro Pipette Connector, COC material, package of 25 piec
  • Microfluidic Micro Fluid Connector, COC material, package of 25 pieces.

Low Flow Pumps

Buy Low Flow Pumps Items Including:
  • Micropump Low Flow pump head, cavity-style design, 0.320 mL/revolution
  • Cole-Parmer Low-Flow Close-Coupled Pump, 316 Stainless Steel, Max Flow (YO-72021-11)
  • Cole-Parmer Low-Flow Close-Coupled Pump, 316 Stainless Steel, Max Flow (YO-72021-12).

Accumet Ph Meter W Stand

Buy Accumet Ph Meter W Stand Items Including:
  • Accumet pH Electrode, Combination, General Purpose, Refillable, Glass
  • Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
  • Oakton Ion 700 Benchtop Meter with "all-in-one" pH electrode and stand (pH/Ion 700) (YO-35419-23).

Lab Supplies Aluminum Sample Cups

Buy Lab Supplies Aluminum Sample Cups Items Including:
  • Medium Lab Frame, Aluminum
  • X-Large Lab Frame, Aluminum
  • Heavy-duty Lab Frame, Aluminum
  • Polystyrene Sample Cups for Flame Photometer Autosampler, 14-mL, 50/pk
  • Sterilized Sampling Bottle Assembly, 125 mL

50 Ml Volumetric Pipette

Buy 50 Ml Volumetric Pipette Items Including:
  • 5.0 mL Unserialized Pipettes (37004-5)
  • 50.0 mL Unserialized Pipettes (37004-50)
  • 50.0 mL Serialized Pipettes (37010-50)
  • 0.5 mL Unserialized Pipettes (37004-510)
  • 1.0 mL Unserialized Pipettes (37004-1)

1000 Ml Separatory Funnel

Buy 1000 Ml Separatory Funnel Items Including:
  • Kimcote Separatory Funnel With Glass Stopper, PTFE, 1000 ml
  • Kimble Separatory Funnel, PTFE Stopcock, 1000 mL 2/cs (YO-34505-50)
  • ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08)
  • Kimble Separatory Funnel, PTFE Stopcock, 2000 mL 2/cs (YO-34505-52).

125 Ml Erlenmeyer Filtering Flask

Buy 125 Ml Erlenmeyer Filtering Flask Items Including:
  • Soxhlet 125 ml round bottom flask (YO-99037-60)
  • KIMAX baffle shake flask, 125 mL 6/cs
  • Thermo Scientific Low Cost Orbital Shaker for 125 mL Erlenmeyer flasks (2345Q)
  • Thermo Scientific Low Cost Orbital Shaker for 125 mL Erlenmeyer flasks (2345-1CEQ).

Lab Stands and Clamps

Buy Lab Stands and Clamps Items Including:
  • Light-duty Converter Clamp for 130-Watt Ultrasonic Processors, fits 1/ (830-00118)
  • Light-duty Converter Clamp for 500- and 750-watt ultrasonic processors (830-00116)
  • Stainless Steel Support Stand Stabilizer Knobs
  • Polycarbonate Lab Jack (183910001).

Ph Meter Oakton Ph 700

Buy Ph Meter Oakton Ph 700 Items Including:
  • Oakton double-junction pH electrode, 10ft cable, BNC connector (YO-35816-77)
  • "All-in-one" pH Electrode DJ with ATC for 600 series meters (YO-35816-72)
  • Cap for Oakton sample bottles, pack of 10
  • Oakton Waterproof pH 600 Meter with pH probe.

Quest 3m Sound Level Meters

Buy Quest 3m Sound Level Meters Items Including:
  • 3M™ SE-401 Sound Level Meter, Type 1
  • 3M™SE-402 Sound Level Meter, Type 2
  • 3M™SE-401-AC3 Sound Level Meter Kit with acoustical calibrator, Typ.

Haake Viscometers

Buy Haake Viscometers Items Including:
  • Thermo Scientific HAAKE Falling Ball Viscometer
  • Thermo Scientific HAAKE Low-range portable viscometer, range 1.5 to 33 (399-0100)
  • Thermo Scientific HAAKE High-range portable viscometer, range 30 to 40 (399-0200).

Clamp Stand

Buy Clamp Stand Items Including:
  • Double Buret Clamp/Stand (916187)
  • Light-duty Converter Clamp for 130-Watt Ultrasonic Processors, fits 1/ (830-00118)
  • Polypropylene Coated Metal Support Stand with Center Rod
  • Cole-Parmer 3feet Device Stand
  • Heavy Duty support stand with 23” s/s rod.

Di Water Conductivity Sensor

Buy Di Water Conductivity Sensor Items Including:
  • GLI 1000 Ohm electrodeless conductivity sensor, PFA, 2" sanitary mount (3708E2T)
  • GLI 1000 Ohm electrodeless conductivity sensor, PP, 3/4 (37 2 5 E2T XX)
  • Cond Twist-Lock Sensor K=0.01
  • Replacement pH/Conductivity/TDS Sensor Module (YO-35425-50).

Oakton Ph Probe Wd 35805 01

Buy Oakton Ph Probe Wd 35805 01 Items Including:
  • Oakton pH electrode, epoxy-body, sealed, double-junction, general purp
  • Oakton PC2700 Meter w/pH electrode, conductivity/temp probe, and elect (YO-35414-01)
  • Oakton PC700 Meter w/pH electrode, conductivity/temp probe, electrode.

5000 Ml Separatory Funnel

Buy 5000 Ml Separatory Funnel Items Including:
  • Kimble Separatory Funnel, PTFE Stopcock, 1000 mL 2/cs (YO-34505-50)
  • Kimble Separatory Funnel, PTFE Stopcock, 2000 mL 2/cs (YO-34505-52)
  • ACE GLASS Separatory Funnels, 1000 mL (YO-34731-08)
  • ACE GLASS Separatory Funnels, 2000 mL (YO-34731-10).



Liquid Sampling to 5000 Ml Separatory Funnel

>