Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 12
Water Pressure Data Logger
to
Luer Lock Fittings Stainless Steel
Image
Product Detail
More Detail
Water Pressure Data Logger
Buy Water Pressure Data Logger Items Including:
Global Water hydrant water pressure logger with 2-1/2" BSPP to 2-1/2" (YO-32922-80)
RHTemp2000 temperature and humidity logger with LCD display (RHTemp2000)
Temperature, Humidity, Pressure, and 50G Tri-axial Shock Recorder (UltraShock-50).
More Details
Tygon Tubing, Fittings, and Hose
Buy Tygon Tubing, Fittings, and Hose Items Including:
Masterflex® Tygon® tubing, nonphthlate/non-DEHP, 1.85mm ID, 1
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16)
3-Stop Tygon® Lab Tubing, 1.14 mm ID, 12/pack (CP95608-30)
3-Stop Tygon® Lab Tubing, 1.52 mm ID, 12/pack (CP95608-36).
More Details
Laboratory Storage Cabinet
Buy Laboratory Storage Cabinet Items Including:
Storage cabinet, 20 drawers (19-320)
Storage cabinet, 16 drawers (19-416)
Storage cabinet, 15 drawers (19-715)
Storage cabinet, 9 drawers (19-909)
Standard cabinet (CV1252TAN)
More Details
Low Flow Micro Water Pump
Buy Low Flow Micro Water Pump Items Including:
Cole-Parmer Low-Flow Close-Coupled Pump, 316 Stainless Steel, Max Flow (YO-72021-11)
Cole-Parmer Low-Flow Close-Coupled Pump, 316 Stainless Steel, Max Flow (YO-72021-12).
More Details
Chloride Meter
Buy Chloride Meter Items Including:
Cole-Parmer Chloride Analyzer, 120v (YO-02656-20)
EM Quant Chloride Test Strips
Chloride Standard, 1 x 100 mL bottle (YO-02656-39)
Chloride (Cl-) ISE Double junction solution kits (YO-27502-63)
ISE Standard solution, chloride, 0.1 M (YO-27503-08).
More Details
Tubing Compatibility Chart
Buy Tubing Compatibility Chart Items Including:
Corrugated FEP Tubing Compatible with Barbed Fittings, 1/4"ID x 9/32"O
Corrugated FEP Tubing Compatible with Barbed Fittings, 3/8"ID x 13/32"
Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft
(YO-96143-02).
More Details
Economy Miniature Peristaltic Pump Head
Buy Economy Miniature Peristaltic Pump Head Items Including:
Masterflex B/T pump head; for IEC 72-71-14F130 motor with B5 flange
Replacement rotor assembly for Masterflex L/S Standard pump heads: 070 (YO-07013-80)
Replacement rotor assembly for Masterflex L/S Standard pump heads: 070 (YO-07013-95).
More Details
Corning Ph Meter 530
Buy Corning Ph Meter 530 Items Including:
Replacement pH electrode, waterproof, for meter kits 58901-00 and -05
pH electrode refillable single-junction for Corning meters (YO-27507-28)
Single-junction (YO-27507-20)
Double-junction (YO-27507-22)
Double-junction with Tuff Tip (YO-27507-24).
More Details
Water Level Sensor Horizontal Float Switch
Buy Water Level Sensor Horizontal Float Switch Items Including:
Standard float switch; polypropylene (solid), 3/8"-16 thread mounting (171514)
Standard float switch; stainless steel with slosh shield, (2) 9/32" mo (43765)
LS-3 Level Switch, Polypropylene Float 0.60 Minimum Liquid Specific Gr (171517).
More Details
Orion Ph Probes
Buy Orion Ph Probes Items Including:
Adapter cable for Orion ATC probes, MiniDIN to 8-pin DIN
Thermo Scientific Orion Stainless Steel Automatic Temperature Compensa
Thermo Scientific Orion 4-Cell Conductivity Probe, K = 0.475, 10-ft ca (013010MD)
Thermo Scientific AquaPro pH Sensor.
More Details
25 Ml Volumetric Flask
Buy 25 Ml Volumetric Flask Items Including:
PYREX Brand 5640 Volumetric Flask; 25 mL, case of 12
PYREX Brand 5580 Volumetric Flask; 25 mL, case of 12
PYREX Brand 5660 Lifetime Red Volumetric Flask; 250 mL, case of 12
PYREX Brand 5660 Lifetime Red Volumetric Flask; 25 mL, case of 12.
More Details
1 Oz Glass Vials
Buy 1 Oz Glass Vials Items Including:
Glass Vials PTFE Lined Cap Clear 2 ml 100/pk (B7800-1)
Boston Round Glass Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs (2300)
Kimble Standard SSr™ Vial, 4ml, 1000/cs (60680-1)
Kimble Straight-sided SSr™ Vial, 4ml, 1000/cs (60690-1).
More Details
Glass Filtering Flask with Side Arm
Buy Glass Filtering Flask with Side Arm Items Including:
KIMAX filtering flask, 25 mL
KIMAX filtering flask, 125 mL
KIMAX filtering flask, 500 mL
KIMAX filtering flask, 1000 mL
Kimcote, Heavy-Wall, Safety-Coated, Filter Flask, 125mL (YO-34524-49)
More Details
Prescription Laboratory Safety Glasses
Buy Prescription Laboratory Safety Glasses Items Including:
Laboratory faceshield, single crown w/acetate shield
Kimcote Volumetric Flask with Glass Stopper; 100mL (YO-34524-31)
Kimcote Volumetric Flask with Glass Stopper; 250mL (YO-34524-33)
Kimcote Volumetric Flask with Glass Stopper; 500mL (YO-34524-35).
More Details
Propeller Mixers
Buy Propeller Mixers Items Including:
316 Stainless Steel three-blade propeller, 2" dia x 3/8" bore dia (YO-04552-55)
Handlebar mixer; 5/8" dia x 30"L shaft, 4" dia three-blade propeller (YO-50500-33)
IKA High-Speed Propeller; Three-Blade, 1.8" (45 mm) dia.
More Details
Clear Plastic Cylinder Containers with Lids
Buy Clear Plastic Cylinder Containers with Lids Items Including:
Hinged-Lid Sample Containers, PP, 1/4 oz, 2500/pk
Hinged-Lid Sample Containers, PP, 1/2 oz, 2000/pk
Hinged-Lid Sample Containers, PP, 2 oz, 750/pk
Hinged-Lid Sample Containers, PP, 4 oz, 300/pk
Attached-lid HDPE containers, 8.5 gal
More Details
Atc Ph Probe
Buy Atc Ph Probe Items Including:
"All-in-One" combination double-junction pH probe with built-in ATC an (YO-05992-16)
Oakton® Acorn™ pH 5 meter with ATC probe (pH 5)
pH/Ion 2100 meter with pH electrode and ATC probe (pH/Ion 2100)
Cole-Parmer Lab-Type ATC Probe, 100Ω RTD.
More Details
Polished Stainless Steel Tubing for Boats
Buy Polished Stainless Steel Tubing for Boats Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Containers Steel
Buy Containers Steel Items Including:
Defenders Stainless Steel Receptacle, 3 1/2 gallon
Stainless steel container with lid 75-500 ML (8520)
Cole-Parmer Covered Container, 304 SS, 1 1/4 qt
Cole-Parmer Covered Container, 304 SS, 2-1/8 qt
Cole-Parmer Covered Container, 304 SS, 4-1/2 qt.
More Details
Uv Lamp 254 Nm
Buy Uv Lamp 254 Nm Items Including:
Standard Pen-Ray UV lamp; 254 nm, 2.13"
Standard Pen-Ray UV lamp; 254 nm, 9"
Battery-operated UV lamp, 254 nm wavelength (95-0009-02)
Standard Pen-Ray UV lamp; 365 nm, 2.25"
20-Watt UV bench lamp; 254 nm, 115 VAC/60 Hz (95-0045-07).
More Details
High Accuracy Thermometer
Buy High Accuracy Thermometer Items Including:
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with genera (1340-5417)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with penetr (1340-5418)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with handle (1340-5416).
More Details
Eppendorf Fixed-Volume 2000 Series Pipettors
Buy Eppendorf Fixed-Volume 2000 Series Pipettors Items Including:
Fixed-volume pipettors, 100 µL (22 47 115-5)
Fixed-volume pipettors, 200 µL (22 47 125-2)
Fixed-volume pipettors, 500 µL (22 47 145-7)
Fixed-volume pipettors, 1000 µL (22 47 165-1)
Fixed-volume pipettors, 1500 µL (22 47 170-8).
More Details
Ertco Temperature Glass Thermometers
Buy Ertco Temperature Glass Thermometers Items Including:
Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (062CBBLSFC)
Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (063FBLSFC)
Thermo Scientific ERTCO® Enclosed Thermometer, range -90 - 20° (15059145).
More Details
Laboratory Hand Pump
Buy Laboratory Hand Pump Items Including:
Complete Laboratory Drum Pump Set, 27" in length, 18.5 GPM, Polypropyl
Complete Laboratory Drum Pumps, 27" in length, 17 GPM, Stainless Steel
Complete Laboratory Drum Pump Set, 27" in length, 17 GPM, Stainless St.
More Details
0.45 Micron Filter Paper Whatman
Buy 0.45 Micron Filter Paper Whatman Items Including:
Whatman Prepleated Filter Papers; 12.5 cm dia; pore size, 8 µ; 100/
Whatman Glass Microfiber Filter Papers Water-Pollution test papers; 4
(YO-06648-70)
Whatman Glass Microfiber Filter Papers Water-Pollution test papers; 4
(YO-06648-71).
More Details
Tubing Pinch Valve
Buy Tubing Pinch Valve Items Including:
Manual Pinch Valve, 5/32" to 1/4" tube OD (PV-2)
Three-way solenoid pinch valve; 12 VDC, 1/8" ID x 1/4" OD tubing (100P3MP-12-05)
Solenoid-Operated Two-Way Pinch Valve; Normally-Open, 10 mm Tube OD, 1 (PK-0802-NO-3 12V N OPEN).
More Details
30 Cm H2o Pressure Gauge
Buy 30 Cm H2o Pressure Gauge Items Including:
Low-Cost Utility Pressure Gauge, 0-30" Hg (KC1012030/0)
Dwyer Magnehelic Differential Pressure Gauge, 2003: 0/3.0 Inh2O
Cole-parmer Industrial Pressure Gauge, 30" Hg To 200 PSI; 1/4" NPT(M)
Ashcroft Pressure Gauge Comparator, 30 psi.
More Details
Nist Traceable Thermometer
Buy Nist Traceable Thermometer Items Including:
NIST-Traceable Pen-Style Pocket Thermometer
Easy-read NIST Certified Thermometer, -10/70°C, 300mm Length, 76mm
Oakton Food TempTestr Infrared Thermometer with NIST-traceable Calibra
Oakton FOOD SAFETY IR THERMOMETER with NIST-traceable calibration.
More Details
Cga 590 Regulator
Buy Cga 590 Regulator Items Including:
Single-Stage Regulator, 2000 scfh Capacity, 590 CGA Fitting (YO-98202-04)
Two-Stage High-Purity Gas Regulator, 270 scfh Capacity, 590 CGA (YO-98210-08)
Two-Stage High-Purity Gas Regulator, 380 scfh Capacity, 590 CGA (YO-98210-18).
More Details
Luer Lock Fittings Stainless Steel
Buy Luer Lock Fittings Stainless Steel Items Including:
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
Compression Straight Union, Stainless Steel, 12mm OD Tubing (YO-31528-36).
More Details
70618
results
<<<Previous
8
9
10
11
12
13
14
15
16
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Water Pressure Data Logger to Luer Lock Fittings Stainless Steel
>
31125329-s-toc_2 31125329stoc2