Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 13

Portable Water Salinity Tester   to   Tygon Microbore Tubing

Image
  Product Detail
More Detail

Portable Water Salinity Tester

Buy Portable Water Salinity Tester Items Including:
  • Extech Conductivity, TDS, Salinity Tester kit
  • Oakton EcoTestr Salt pocket salinity tester
  • Extech EC410 conductivity/TDS/salinity tester kit
  • Cole-Parmer® Salinity Pocket Tester
  • Oakton SALT 6+ handheld salinity meter with probe and NIST-traceable C.

Coorstek Crucibles

Buy Coorstek Crucibles Items Including:
  • CoorsTek Crucible Cover, Porcelain; 26 mm ID, for 17803-02, pk of 12
  • CoorsTek Crucible Cover, Porcelain; 34 mm ID, for 17803-04, 17804-02, (60122) (YO-17811-02)
  • CoorsTek Crucible Cover, Porcelain; 26 mm ID, for 17803-02, cs of 72.

Ssi Digital Pressure Gauge

Buy Ssi Digital Pressure Gauge Items Including:
  • Digital Pressure Gauge, 0-100 PSI
  • Digital Pressure Gauge, 0-200 PSI
  • Digital Pressure Gauge, 0-300 PSI
  • Digital Pressure Gauge, 0-500 PSI
  • Digital Pressure Gauge, 0-1000 PSI

Chemistry Supplies Mortar and Pestle

Buy Chemistry Supplies Mortar and Pestle Items Including:
  • Cole-Parmer Mortar and Pestle Set, Porcelain; 80ML, Each
  • Cole-Parmer Mortar and Pestle Set, Porcelain; 500ML Each
  • Mortar and Pestle; Agate, 125ML, Each
  • Agate Mortar and Pestle, 25 mL (YO-04010-20)
  • Glass Mortar & Pestle Set, 50ml (YO-34500-51).

3-Way Solenoid Pinch Valves

Buy 3-Way Solenoid Pinch Valves Items Including:
  • Three-way solenoid pinch valve; 12 VDC, 1/8" ID x 1/4" OD tubing (100P3MP-12-05)
  • Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 13 mm Tube OD, (EPK-1005-NC 24V N CLOSED).

Portable Bod Meter

Buy Portable Bod Meter Items Including:
  • Thermo Scientific Orion Star A223 DO Portable Meter Only
  • Thermo Scientific Orion Star A123 Dissolved Oxygen Portable Meter
  • Thermo Scientific Orion Star A123 Dissolved Oxygen Portable Meter Kit.

55 Gallon Drum Rotary Hand Pump

Buy 55 Gallon Drum Rotary Hand Pump Items Including:
  • 12in Threaded Standoff Kit For 55 Gallon Drums
  • Drip Sentinel Drum-top Workstation For 55 Gallon Steel Drums
  • Hand-operated threaded hand pump, 26 strokes/gallon, 3/4" NPT(F) adapt (MP1209)
  • Guzzler diaphragm hand pump, 8.5 GPM, 7 Strokes/gallon.

Gas Flow Meter Pulse Output

Buy Gas Flow Meter Pulse Output Items Including:
  • pH pulse output controller
  • Magnetic Low-Flow Flowsensor, 0.2-20 GPM, Pulse and 4 to 20 mA Output (YO-33111-51)
  • Pulse input rate/totalizer, two SPDT relays and 4 to 20 mA output, 85
  • Pulse input rate/totalizer, 4 to 20 mA output, 85 to 265 VAC power.

Seripettor Bottletop Dispenser 2.5 25ml

Buy Seripettor Bottletop Dispenser 2.5 25ml Items Including:
  • Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
  • Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Glass Bottle
  • Cole-Parmer Bottletop Dispenser 2.5-30 mL
  • Replacement Cartridges for BrandTech Seripettor Economy Dispenser, 2.5.

Polyethylene Tubing Chemical Resistance

Buy Polyethylene Tubing Chemical Resistance Items Including:
  • Tygon 2375 Ultra Chemical-Resistant Tubing, 1/16"ID x 1/8"OD, 50 ft/pa
  • Tygon 2375 Ultra Chemical-Resistant Tubing, 1/16"ID x 3/16"OD, 50 ft/p
  • Tygon 2375 Ultra Chemical-Resistant Tubing, 3/32"ID x 5/32"OD, 50 ft/p.

Uv Light Meter Reviews

Buy Uv Light Meter Reviews Items Including:
  • UV Intensity Meter Sensor, 365 nm (97-0016-02(UVX36))
  • UV Intensity Meter Sensor, 302 nm (97-0016-04(UVX31))
  • J-225:Uv Meteranalog Short Wave 97-0004-01 (97-0004-01)
  • Short/long-wave UV lamp; 4 watts, 254/365 nm wavelength, 115 VAC/60 Hz (95-0021-12).

Horn Sonicator

Buy Horn Sonicator Items Including:
  • Cup horn (3") for use with ultrasonic processors
  • Cup horn (5.5") for use with ultrasonic processors
  • Dual Horn (with two 3/4" solid probes) for use with ultrasonic process
  • Dual Horn (without probes) for use with ultrasonic processors.

Oakton Ph Tester 10

Buy Oakton Ph Tester 10 Items Including:
  • Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
  • Oakton Waterproof pHTestr 10 Pocket pH Tester and calibration kit
  • Oakton Waterproof pHTestr 20 Pocket pH Tester and calibration kit
  • Oakton Waterproof pHTestr 30 Pocket pH Tester and calibration kit.

Portable Dissolved Oxygen Meter Orbisphere A1100

Buy Portable Dissolved Oxygen Meter Orbisphere A1100 Items Including:
  • Thermo Scientific Orion Star A123 Dissolved Oxygen Portable Meter Kit
  • Thermo Scientific Orion Star A223 DO Portable Meter Kit
  • Thermo Scientific Orion Star A223 DO Portable Meter Only
  • Thermo Scientific Orion Star A323 RDO/DO Portable Meter Kit.

Micro Flow 3 Way Valves

Buy Micro Flow 3 Way Valves Items Including:
  • Cole-Parmer High-Flow Two-Way Solenoid Valve for Steam, 3/4" Orifice S
  • Hayward electrically Actuated Ball Valve, 3-way, 3/4" PVC (HCTN1075STV/EAU-28)
  • Hayward electrically Actuated Ball Valve, 3-way, 3/4" CPVC (HCTN2075STV/EAU-28).

Lab Spill Kit

Buy Lab Spill Kit Items Including:
  • Hazardous lab spill clean-up kit
  • Laboratory Safety Spill Kit; 3.5 Gal.; 1 Pack/case
  • Lab & Cleanroom Spill Station.

Thin Walled Stainless Steel Tubing

Buy Thin Walled Stainless Steel Tubing Items Including:
  • 316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
  • 304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
  • Stainless steel tube set for electric and air motors, 27"L.

Oakton Pc 650

Buy Oakton Pc 650 Items Including:
  • Oakton Waterproof PC 650 Meter with probes
  • Oakton Waterproof PC 650 Meter with probes and NIST-traceable Calibrat
  • Oakton Waterproof PC 650 Meter only with NIST-traceble Calibration
  • Oakton Waterproof PC 650 Meter Kit with NIST-traceable Calibration.

Cole-Parmer Specific Gravity Cups

Buy Cole-Parmer Specific Gravity Cups Items Including:
  • Specific Gravity Cup, 83.2 mL capacity, 0.5% volume tolerance, for use (PV-9654)
  • Specific gravity cup, 8.32 mL capacity, 1.2% volume tolerance, for use (WG-SS-8.32)
  • Specific gravity cup, 83.2 mL capacity, 0.5% volume tolerance, for use (WG-SS-83.2).

Drying Rack Stainless

Buy Drying Rack Stainless Items Including:
  • Modular stainless steel drying rack, 32 assorted color pegs
  • Modular stainless steel drying rack, 50 assorted color pegs
  • Modular stainless steel drying rack, 60 assorted color pegs
  • Drain basket for modular stainless steel drying racks.

Dry Ice Storage Containers

Buy Dry Ice Storage Containers Items Including:
  • Dry ice storage chest, 3.75 cu ft (YO-03720-00)
  • Dry ice storage chest, 2.5 cu ft (YO-03721-00)
  • Dry ice storage chest, 1.6 cu ft (YO-03721-02)
  • Ice storage chest (YO-03719-00)
  • DOLLY F/03720-00 03745-00 (376).

Low Flow Pumps Submersible

Buy Low Flow Pumps Submersible Items Including:
  • Micropump Low Flow pump head, cavity-style design, 0.320 mL/revolution
  • Variable speed drive for Micropump low-flow pump heads, 6000 rpm, 0 to
  • Variable speed drive for Micropump low-flow pump heads, 8000 rpm, 0 to.

Costar Plates

Buy Costar Plates Items Including:
  • Storage Mat for Costar 96-Well Polypropylene Storage System
  • Costar 96-Well Standard Microplates, Nontreated, PP, Flat Well
  • Costar 96-Well Standard Microplates, Nontreated, PP, Round Well
  • Costar 96-Well Assay Blocks, Nontreated, PP, 1.0 mL, Sterile.

Lab Bottle Top Dispenser

Buy Lab Bottle Top Dispenser Items Including:
  • Bottle top adapter (TZ 2005)
  • Cole-Parmer Bottletop Dispenser .25-2.5 mL
  • Cole-Parmer Bottletop Dispenser 2.5-30 mL
  • Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
  • Cole-Parmer Bottletop Dispenser .5-5 mL with 100 mL Glass Bottle.

Clear Glass Vial with Screw Cap

Buy Clear Glass Vial with Screw Cap Items Including:
  • Glass Vials PTFE Lined Cap Clear 12 ml 200/pk (B7800-12)
  • Glass Vials PTFE Lined Cap Clear 16 ml 200/pk (B7800-4)
  • Glass Vials PTFE Lined Cap Clear 20 ml 100/pk (B7800-20)
  • Glass Vials PTFE Lined Cap Clear 22 ml 200/pk (B7800-5).

Non Glass Ph Probe

Buy Non Glass Ph Probe Items Including:
  • Honeywell Durafet III, non-glass pH electrode, 3/4 inch immersion thre
  • Honeywell® Durafet III, non-glass pH electrode, 2.0" Tri-Clamp fla (07794DVP-04-02)
  • Honeywell® Durafet III, non-glass pH electrode, 3.0" Tri-Clamp fla (07794DVP-06-02).

20 Mm Diameter Petri Dish

Buy 20 Mm Diameter Petri Dish Items Including:
  • Petri dish with pads, 50x11 mm, 100/cs (YO-14005-20)
  • Petri dish without pads, 50x11 mm, 100/cs
  • Petri dish without pads, 50x11 mm, 500/cs
  • Petri dish with pads, 50x11 mm, 500/cs
  • Thermo Scientific Nalgene Autoclavable Petri Dishes, PMP, 100 x 15 mm,.

Thermo Scientific Labquake Shakers

Buy Thermo Scientific Labquake Shakers Items Including:
  • Thermo Scientific Labquake Tube Shaker/Rotators, Large, 14 Tubes, 50 m (4152110Q)
  • Thermo Scientific Labquake Tube Shaker/Rotators, Large, 14 Tubes, 50 m (4152220Q)
  • Thermo Scientific Labquake Tube Shaker/Rotators, Large, 46 Tubes, 10 m (T415110Q).

1.3125 Diameter Stainless Steel Tubing

Buy 1.3125 Diameter Stainless Steel Tubing Items Including:
  • 316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
  • 304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
  • Top Cover with Handle for 12" Diameter Stainless Steel Sieve.

Tygon Microbore Tubing

Buy Tygon Microbore Tubing Items Including:
  • Tygon Microbore Tubing, 0.010" x 0.030"OD, 100 ft/roll
  • 3-Stop Tygon MHLL Tubing, 0.76 mm ID, 6/pack
  • 3-Stop Tygon MHLL Tubing, 1.14 mm ID, 6/pack
  • 3-Stop Tygon MHLL Tubing, 1.52 mm ID, 6/pack
  • 3-Stop Tygon MHLL Tubing, 2.03 mm ID, 6/pack.



Portable Water Salinity Tester to Tygon Microbore Tubing

>