Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 14
Kynar Pvdf Tubing
to
Chloride Ion Selective Electrode
Image
Product Detail
More Detail
Kynar Pvdf Tubing
Buy Kynar Pvdf Tubing Items Including:
KynarSuperflex PVDF Tubing 1/16" x 1/8" 25ft /pk
KynarSuperflex PVDF Tubing - 3/16" x 1/4" 25 ft/pk
KynarSuperflex PVDF Tubing 1/4" x 3/8" 25 feet/pk
Tubing Adapter; Kynar, 1/2" mini to 1/8" NPT (M).
More Details
Vacuum Pump Regulator
Buy Vacuum Pump Regulator Items Including:
Vacuum/pressure Diaphragm Pump With Gauges And Regulators, PTFE-coated (107CA18TFEL-246)
Vacuum/pressure diaphragm pump with gauges and regulators, PTFE-coated (107CDC18TFEL-246)
Vacuum Regulator, 0 to 30" Hg, 2 cfm Max Capacity (AJ675).
More Details
Silastic Tubing
Buy Silastic Tubing Items Including:
Silastic Laboratory Tubing, .012" ID x .025" OD, 50'.
More Details
Glass Syringes
Buy Glass Syringes Items Including:
Cadence Science® Micro-Mate 20 cc syringe with metal luer lock tip
Cadence Science® Micro-Mate 10 cc syringe with metal luer slip tip
Cadence Science® Micro-Mate 10 cc syringe with glass luer slip tip.
More Details
Lab Jack
Buy Lab Jack Items Including:
Jiffy-Jack Lab Jack, 4 x 4” Aluminum Lab-Lift
Jiffy-Jack Lab Jack, 6 x 6” Aluminum Lab-Lift
Jiffy-Jack Lab Jack, 8 x 8” Aluminum Lab-Lift
Jiffy-Jack Lab Jack, 66 lb/30 kg limit
Jiffy-Jack Lab Jack, Heavy-Duty Apparatus Positioners, 80lb (136.3kg).
More Details
Accumet Ph Electrode
Buy Accumet Ph Electrode Items Including:
accumet epoxy body, non-temperature compensated, 2-cell electrode, K= (YO-19550-60)
accumet epoxy body, non-temperature compensated, 2-cell electrode, K= (YO-19550-62)
accumet epoxy body, temperature compensated, 2-cell electrode, K=1 (YO-19550-64).
More Details
Carbon Dioxide Incubator Gas
Buy Carbon Dioxide Incubator Gas Items Including:
Carbon Dioxide Traps; 40 liters volume; 1/8" fittings (YO-34651-60)
Carbon Dioxide Traps; 40 liters volume; 1/4" fittings (YO-34651-70)
Sensidyne Gas Detection Tubes, Carbon dioxide
Sensidyne Gas Detection Tubes, Carbon Dioxide, 5 to 50%.
More Details
Art Pipette Tips
Buy Art Pipette Tips Items Including:
Tip Art .1-10µL Sterile 960/pk (2139-05)
Tip Art .1-10µL Sterile 960/pk (2140-05)
Tip Art .1-10µL Sterile 960/pk (2149E-05)
Tip Art .1-10µL Sterile 960/pk (2149E)
Tip Art 0.5-20µL Sterile 960/pk (2149P-05)
More Details
Hotplate Stirrer with External Temperature Probe
Buy Hotplate Stirrer with External Temperature Probe Items Including:
Cole-Parmer StableTemp Dry Block Heaters External Temperature Probe Ki
External temperature probe for MicroLog and MicroLogPROII Data Loggers
VAISALA RH-temperature transmitter w/Display, 3 Meter Remote Probe and.
More Details
Burkert Solenoid Valve Distributors
Buy Burkert Solenoid Valve Distributors Items Including:
Cole-Parmer Two-Way Micro Solenoid Rocker Valve, 1/4-28 UNF
Cole-Parmer Two-Way Micro Solenoid Rocker Valve, Manifold Mount
Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 1/4-28 UNF
Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 1/8 NPT(F).
More Details
Soil Sieve Set Stainless Steel
Buy Soil Sieve Set Stainless Steel Items Including:
Receiving Pan for 12" Stainless Steel Bottom Sieves, Full Height
Receiving Pan with Fitted Rim for Nesting 12" Stainless Steel Sieves, (YO-59996-92)
Receiving Pan with Fitted Rim for Nesting 12" Stainless Steel Sieves, (YO-59996-93).
More Details
Nylon Mesh Filter Screen Seattle
Buy Nylon Mesh Filter Screen Seattle Items Including:
Millipore Steriflip Vacuum-Driven Filter Device with Nylon mesh membra
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 100 µm, 3/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 70 µm, 3/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 60 µm, 3/pk.
More Details
Cryogenic Dewars
Buy Cryogenic Dewars Items Including:
Cole-Parmer Aluminum - Glass Dewar Flask with Handle, 1200 mL
Cole-Parmer Aluminum - Glass Dewar Flask with Handle, 1500 mL
Cole-Parmer Aluminum - Glass Dewar Flask with Handle, 2000 mL
Cole-Parmer Aluminum - Glass Dewar Flask with Handle, 2500 mL.
More Details
Food Grade 316 Stainless Steel Tubing
Buy Food Grade 316 Stainless Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
316 Stainless Steel fittings, female luer x female luer (YO-31507-89)
316 Stainless Steel fittings, male luer lock x male luer lock (YO-31507-90).
More Details
Oakton Ph Calibration Standards
Buy Oakton Ph Calibration Standards Items Including:
Oakton pH 5+ meter kit with NIST-traceable Calibration
Oakton pH 6+ meter kit with NIST-traceable Calibration
Oakton pH 11 meter kit with NIST-traceable Calibration
Oakton pH buffer pouches; pH 4.01, 20/pk
Oakton pH buffer pouches; pH 7.00, 20/pk.
More Details
2 Way Plastic Luer Stopcock
Buy 2 Way Plastic Luer Stopcock Items Including:
Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pac (YO-31200-90)
Luer stopcock, 1-way, male luer lock x female luer, black PVDF, pack o (YO-31200-91)
PTFE 2-Way Stopcock, 1/2" Orifice, 1/2" Tube compression connections.
More Details
Marsh Bellofram Type 1000 I/P Transducers
Buy Marsh Bellofram Type 1000 I/P Transducers Items Including:
Pressure Transducer, I to P;3-15 PSI Output, 4-20 mA Input (961-070-000)
Pressure Transducer, I to P;3-27 PSI Output, 4-20 mA Input (961-074-000)
Pressure Transducer, I to P;6-30 PSI Output, 4-20 mA Input (961-075-000).
More Details
Sanitary Reducer Fitting
Buy Sanitary Reducer Fitting Items Including:
316L Stainless Steel Asme-bpe Sanitary Clamp Reducing Tee, 20ra Mechan (YO-30606-19)
316L Stainless Steel Asme-bpe Sanitary Clamp Reducing Tee, 20ra Mechan (YO-30606-20).
More Details
Actuated Ball Valve Pvc
Buy Actuated Ball Valve Pvc Items Including:
Hayward electrically Actuated Ball Valve, 3-way, 1" PVC (HCTN1100STV/EAU-28)
Hayward electrically Actuated Ball Valve, 3-way, 2" PVC (HCTN1200STV/EAU-28)
Hayward electrically Actuated Ball Valve, 3-way, 1/2" CPVC (HCTN2050STV/EAU-28).
More Details
Thermo Scientific Universal Repipet Dispensers
Buy Thermo Scientific Universal Repipet Dispensers Items Including:
Thermo Scientific Nalgene PPCO dispensing bottle, 125 mL
Thermo Scientific Nalgene PPCO dispensing bottle, 250 mL
Thermo Scientific Finnpipette ® Dispenser, 0.2 to 1.0 mL (4421120)
Thermo Scientific Finnpipette ® Dispenser, 0.4 to 2.0 mL (4421130).
More Details
Oakton Thermometer
Buy Oakton Thermometer Items Including:
Oakton® Infrared Thermometer with Type-K Input (High Temp) (YO-39644-10)
Oakton Temp-16 Precision RTD Thermometer w/boot (CPTEMP16)
Oakton Temp-10 type J thermocouple thermometer with boot
OAKTON Temp-10 type K thermocouple thermometer with boot.
More Details
Water Grab Sampler
Buy Water Grab Sampler Items Including:
Open water grab sampler, 6' long
Open water grab sampler, 12' long
Open water grab sampler, 18' long
Replacement bottle/cap for open water grab sampler
Ponar grab dredge, 6" x 6" sampling area.
More Details
Pvc Coated Stainless Steel Tubing
Buy Pvc Coated Stainless Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Swagelok Bulkhead Compression Fitting
Buy Swagelok Bulkhead Compression Fitting Items Including:
Compression fittings, Bulkhead union, Kynar, 1/4" OD, 2-9/32", 5/8", 7 (YO-30805-16)
Compression Fitting, Bulkhead Union, 316 SS, 1/8" OD (YO-31406-12)
Compression Fitting, Bulkhead Union, 316 SS, 1/4" OD (YO-31406-13).
More Details
Oakton Ph 11 Series
Buy Oakton Ph 11 Series Items Including:
Oakton pH 11 meter with "All-in-One" electrode (35811-71)
Oakton pH 11 meter only with NIST-traceable Calibration
Oakton pH 11 meter kit with NIST-traceable Calibration
Oakton ECTestr 11+ multi-range, cup-style pocket conductivity tester.
More Details
Oakton Ph 5 Acorn Series
Buy Oakton Ph 5 Acorn Series Items Including:
Oakton pH 5+ meter with probe
Oakton pH 5+ meter kit
Oakton pH 5+ meter with probe with NIST-traceable Calibration
Oakton Acorn pH 5 meter with electrode, and ATC probe (pH 5)
Oakton ATC probe for pH 5, pH 6, Ion 6, pH 11, pH 110, pH 510, Ion 510.
More Details
Coors Buchner Funnel no 60248 4500ml
Buy Coors Buchner Funnel no 60248 4500ml Items Including:
Buchner funnel mantle for coors funnel no
60248, 350 watts, 230 VAC (100AO823)
Buchner funnel mantle for coors funnel no
60248, 350 watts, 115 VAC (100AO822)
Buchner funnel mantle for coors funnel no
More Details
Pyrex Tubing
Buy Pyrex Tubing Items Including:
Pyrex Brand 9800 test tube; 3 mL, pack of 72
Pyrex Brand 9825 culture tube with screw cap; 15 mL, pack of 144
Pyrex Brand 9825 culture tube with screw cap; 20 mL, pack of 144
Pyrex Brand 9825 culture tube with screw cap; 25 mL, pack of 96.
More Details
Size 16 Pharmed Tubing
Buy Size 16 Pharmed Tubing Items Including:
Thermo Scientific Nalgene Snap-On Polypropylene Closures; size 16; 20/
PharMed BPT Tubing,3/8x5/8",25'
PharMed BPT Tubing,3/16x5/16",25'
PharMed BPT Tubing,5/16x7/16",25'
Ace Glass Tube Adapter with Hose Barb, 24/40 Joint (5261-16).
More Details
Chloride Ion Selective Electrode
Buy Chloride Ion Selective Electrode Items Including:
Cole-Parmer® Combination Ion Selective Electrodes, Chloride (Cl) (YO-27504-08)
ISE Standard solution, chloride, 0.1 M (YO-27503-08)
Thermo Scientific Orion ISE chloride electrode, solid-state, BNC conne (9417BN)
SOLUTION KIT CHLORIDE (YO-35802-62).
More Details
70618
results
<<<Previous
10
11
12
13
14
15
16
17
18
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Kynar Pvdf Tubing to Chloride Ion Selective Electrode
>
31125329-s-toc_2 31125329stoc2