Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 16
Polyethylene Rigid Tubing
to
Dry Heating Block
Image
Product Detail
More Detail
Polyethylene Rigid Tubing
Buy Polyethylene Rigid Tubing Items Including:
High-Density Polyethylene (HDPE) Tubing, 0.17 x 1/4", 100 ft/pack
High-Density Polyethylene (HDPE) Tubing, 3/16 x 5/16", 100 ft/pack
High-Density Polyethylene (HDPE) Tubing, 5/16 x 7/16", 100 ft/pack.
More Details
Pall Reusable Syringe Filter Holders
Buy Pall Reusable Syringe Filter Holders Items Including:
Gasket,PTFE; for stainless steel syringe filter holder, 13 mm
Advantec Stainless Steel Filter Holder, Syringe; 13mm dia
Advantec Stainless Steel Filter Holder, Syringe; 25mm dia
Advantec High-Pressure Stainless Steel Filter Holder, 47 mm.
More Details
Stainless Steel Junction Boxes
Buy Stainless Steel Junction Boxes Items Including:
12x10x6 Type 304 Junction Box, Stainless Steel (JB121006H4)
12x10x6 Type 316L Junction Box, Stainless Steel (JB121006H6)
12x12x6 Type 316L Junction Box, Stainless Steel (JB121206H6)
14x12x6 Type 316L Junction Box, Stainless Steel (JB141206H6).
More Details
Tall Narrow Glass Beakers
Buy Tall Narrow Glass Beakers Items Including:
Borosil, Beaker, 500ml/6pk (BG1060-500)
Pyrex Brand 1060 Berzelius Tall Form Beaker, 100 mL, 12/pk (1060-100)
Pyrex Brand 1060 Berzelius Tall Form Beaker, 400 mL, 6/pk (1060-400)
Pyrex Brand 1060 Berzelius Tall Form Beaker, 500 mL, 6/pk (1060-500).
More Details
Specific Gravity Hydrometers
Buy Specific Gravity Hydrometers Items Including:
H-B Instrument Specific Gravity Hydrometer Set, 8 Plain Form, 0.700 to
Durac Baume & Specific Gravity Hydrometer, 1.800/2.000 Sp Gr, 0.005 Di
Durac Specific Gravity Hydrometer, 0.600 To 0.670, 0.001 Div, 165mm L,.
More Details
Corning Micro Ph Electrode
Buy Corning Micro Ph Electrode Items Including:
Three-in-one pH/reference/temperature electrode, epoxy body, BNC for m (476436)
Thermo Scientific Orion pH microelectrode, glass body, refillable, sin (9826BN)
Three-in-one pH/reference/temperature electrode, epoxy body, BNC for m (476436P).
More Details
0.45 Um Glass Fiber Syringe Filters
Buy 0.45 Um Glass Fiber Syringe Filters Items Including:
DynaGard Hollow Fiber Syringe Filter for Aqueous Solutions, 0.2 um Por (YO-29510-70)
DynaGard Hollow Fiber Syringe Filter for Aqueous Solutions, 0.2 um Por (YO-29510-72)
Cole-Parmer HPLC Syringe Filters, Non Sterile, PTFE, 0.45 um, 4 mm dia.
More Details
200 Ml Corex Glass Centrifuge Tubes
Buy 200 Ml Corex Glass Centrifuge Tubes Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
10ml Glass Centrifuge Tubes
Buy 10ml Glass Centrifuge Tubes Items Including:
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 5 ml, 125 (99502-5)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 10 ml, 12 (99502-10)
Conical-Bottom Disposable Glass Centrifuge Tubes, Screw Cap, 15 ml, 12 (99502-15).
More Details
Oakton Ph Singles
Buy Oakton Ph Singles Items Including:
Oakton pH 300 meter with single-junction pH/ATC probe
Oakton pH 310 meter with single-junction pH/ATC probe with NIST-tracea
Oakton pH 300 meter with single-junction pH/ATC probe with NIST-tracea
Oakton® Acorn™ TDS 6 Meter with probe (YO-35606-15).
More Details
Digi Sense Rtd
Buy Digi Sense Rtd Items Including:
Digi-Sense® Standard Temperature Controller, 110V (CPTEMCTPSTD01)
Digi-Sense® Standard Temperature Controller, 230V (CPTEMCTPSTD02)
Digi-Sense® Advanced Temperature Controller, 115V (CPTEMCTPDLX01)
Cole-Parmer 100 ohm Platinum RTD Probes; 5"L.
More Details
Ross Electrode Filling Solution Msds
Buy Ross Electrode Filling Solution Msds Items Including:
ROSS electrode fill solution, 3 M KCl
Case of five 60-mL bottles (810007)
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Filling solutions: use with Cl- electrodes (900017)
Filling solution f/ammonia ISE electrode (951202).
More Details
50 L Carboy 3 Sanitary
Buy 50 L Carboy 3 Sanitary Items Including:
High-Purity Liquid Centrifugal Pump, 316L Stainless Steel, 3 Hp, 12 GP.
More Details
Tubing Heater
Buy Tubing Heater Items Including:
Thermo Scientific Heating block for 6 mm tubes, 30 openings
Thermo Scientific Heating block for 8 mm tubes, 30 openings
Thermo Scientific Heating block for 10 mm tubes, 20 openings
Thermo Scientific Heating block for 12 or 13 mm tubes, 22 openings.
More Details
100 Ft Metal Capillary Tube
Buy 100 Ft Metal Capillary Tube Items Including:
Low-Density Polyethylene (LDPE) Tubing, 5/64" x 1/8", 100 ft/bag
Low-Density Polyethylene (LDPE) Tubing, 7/64" x 5/32", 100 ft/bag
Low-Density Polyethylene (LDPE) Tubing, 1/8" x 3/16", 100 ft/bag.
More Details
Handheld Vacuum Gauge Inches Water
Buy Handheld Vacuum Gauge Inches Water Items Including:
Handheld Vacuum Meter, 0-1999 mbar
Fixed Snubber, Brass, 1500 psi, Water or Light Oil
Fixed Snubber, 304/316 SS, 5000 psi, Water or Light Oil
-100 To 100 PSId Handheld Portable Differential Gauge
-5 To 5 PSId Handheld Differential Pressure Gauges.
More Details
2 Ml Micro Burette
Buy 2 Ml Micro Burette Items Including:
Gilmont micrometer burette, 2 mL
Sample Vial Holder for Carbon Residue Tester, seven places for 2-mL an
Borosilicate Glass Vial, 2 mL, pack of 144
Quartz Reusable Vial, 2 mL
Self-masking micro cuvette; cover top; quartz; 0.7 mL capacity, 2/pack.
More Details
Vacuum Pump Filters
Buy Vacuum Pump Filters Items Including:
Filter Pack For 35031-00 & -10 Vacuum Pumps, 10 Pack (72210)
Inlet/Outlet filter, 10 micron filter, 31.8 CFM, 1/2" NPT(F)
Inlet/Outlet filter, 10 micron filter, 180 CFM, 1 1/2" NPT(F)
Oil mist filter, 4.5 cfm, 1/2" NPT(M).
More Details
Alnor Anemometers
Buy Alnor Anemometers Items Including:
TSI™ Alnor CompuFlow Thermoanemometer, manual datalogging
TSI™ Alnor CompuFlow Thermoanemometer, auto datalogging w/humidity
Alnor RVA801 Hand-held vane anemometer
Alnor RVA501 : Data Logging Vane Anemometer Hand Held.
More Details
Bspt to Npt Adapter
Buy Bspt to Npt Adapter Items Including:
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPT (YO-31830-40)
Threaded adapter, stainless steel, 1/4" female NPT to 1/4" male BSPT (YO-31830-41).
More Details
Steam Stainless Steel Immersion Heaters
Buy Steam Stainless Steel Immersion Heaters Items Including:
Immersion heater, short rod, 5.5" L heated area, 500 watts, 115 VAC (324-114)
Immersion heater, short rod, 5.5" L heated area, 500 watts, 230 VAC (306-114)
Immersion heater, loop, 3-1/8" dia
heated area, 500 watts, 115 VAC (335-115).
More Details
Wireless Refrigerator Freezer Thermometer with Alarm
Buy Wireless Refrigerator Freezer Thermometer with Alarm Items Including:
Digital-Bottle Refrigerator/Freezer Thermometer with solution filled b
Digital-Bottle High Accuracy Refrigerator/Freezer Thermometer with sol
Jumbo Refrigerator/freezer thermometer with a 5ml bottle that fits in (4627CP).
More Details
Pyrex Beaker 500 Ml
Buy Pyrex Beaker 500 Ml Items Including:
Graduated polypropylene beaker, 500 mL (16 oz), 25 pk
Pyrex Large Vol Burette 500 ml (2094-500)
Pyrex Brand 6402 Separatory Funnel; 500 mL, pack of 1
Pyrex Brand 6402 Separatory Funnel; 500 mL, case of 4
PYREX Brand 4980 flask; 500 mL, case of 36.
More Details
Combination Electrodes Glass Body Accumet
Buy Combination Electrodes Glass Body Accumet Items Including:
Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
Accumet Accutuph Plus pH Electrode, Combination, Refillable, Glass Bod
Accumet Accutuph pH Electrode, Combination, Single-junction, Refillabl.
More Details
Vacuum Sample Containers
Buy Vacuum Sample Containers Items Including:
Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 30 mL, 20 mm Ne
Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 60 mL, 20 mm Ne
Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 120 mL, 22 mm N.
More Details
Turbidity Meter Calibration Standard
Buy Turbidity Meter Calibration Standard Items Including:
Calibration standards for Hach turbidity meter, item number 99511-00 (26594-05)
HF scientific Solid Validation Turbidity Standard, Low NTU
HF scientific Solid Validation Turbidity Standard, Mid NTU
Hach StablCal 1000 NTU Turbidity Standard, 500-mL.
More Details
Small Battery Operated Magnetic Stirrer
Buy Small Battery Operated Magnetic Stirrer Items Including:
Cole-Parmer Battery-Powered Magnetic Stirrer
Support Stand for IKA RCT and RET Basic Stirrers (2636700)
Ika Ultra-Flat Lab Disc Stirrer 230V (3277700)
Ika Ultra-Flat Lab Disc Stirrer Orange (3578000)
Ika Ultra-Flat Lab Disc Stirrer Pink (3578400).
More Details
Polyethylene Tubing for Instrument Air Supply
Buy Polyethylene Tubing for Instrument Air Supply Items Including:
Flexelene SFX Flexible Polyethylene Tubing, 1/8 x 1/4, 100' per pack (YO-96180-03)
Flexelene SFX Flexible Polyethylene Tubing, 1/4 x 3/8, 100' per pack (YO-96180-05)
Flexelene SFX Flexible Polyethylene Tubing, 3/8 x 1/2, 100' per pack (YO-96180-07).
More Details
Gilson Pipettes 20 200
Buy Gilson Pipettes 20 200 Items Including:
Labpette Discovery Pipette 20 - 200 uL (P3940-200)
Cole-Parmer Adjustable Volume Pipette 20 to 200 µL
Ovation Adjustable-Volume Pipette, 20 to 200 µL, Blue
Single-Channel Adjustable-Volume 20-200µL (W810312)
EcoPipette Variable 20-200 ul (C200-1).
More Details
Dry Heating Block
Buy Dry Heating Block Items Including:
Thermo Scientific Direct contact microtiter heating block
Cole-Parmer StableTemp Single Block for Conical Bottom Centrifuge Tube (949127)
Cole-Parmer StableTemp Single Block for PCR, 10 strips or 8 tubes
Cole-Parmer StableTemp Single Block Cuvette.
More Details
70618
results
<<<Previous
12
13
14
15
16
17
18
19
20
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Polyethylene Rigid Tubing to Dry Heating Block
>
31125329-s-toc_2 31125329stoc2