Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 17
Air Products Regulators
to
Stainless Steel
Image
Product Detail
More Detail
Air Products Regulators
Buy Air Products Regulators Items Including:
Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 (960-115-000)
Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 (960-116-000)
Marsh Bellofram Type 10 Pressure Regulator, 1/8" NPT(F), 2-25 psi.
More Details
10 Ml Glass Pipette
Buy 10 Ml Glass Pipette Items Including:
Thermo Scientific Finntip 10 mL Sterile Pipette Tips, 2 to 10 mL, 5x24
1.0 mL Unserialized Pipettes (37004-1)
Pyrex 7077 Pipette, 10 ML, 600/CS
Brand Positive Displacement Pipettor Tips, PP, 2-10 mL, 10/pk
Disposable Tips, 2-10 mL, 200/bg (BMT-Z).
More Details
Norprene Hose
Buy Norprene Hose Items Including:
Norprene food tubing, 1/16"ID x 3/16"OD, 50 ft/pack
Norprene food tubing, 1/8"ID x 1/4"OD, 50 ft/pack
Norprene food tubing, 3/16"ID x 5/16"OD, 50 ft/pack
Norprene pressure tubing, 1/4"ID x 1/2"OD, 50 ft/pack.
More Details
Battery Operated Vacuum Pumps
Buy Battery Operated Vacuum Pumps Items Including:
Pump, Drum Pump, Rechrageable Battery Operated, Polypropylene, 19"
Repair kit for vacuum/pressure pump 79301-11
12 VDC Rechargeable Gel Battery for Global Water Fluid Samplers (00-010)
120 VAC to 12 VDC Battery Charger for 99243-07 (FE0400).
More Details
Dissolved Oxygen Test Kit Test Kits
Buy Dissolved Oxygen Test Kit Test Kits Items Including:
Hach Dissolved Oxygen Test Kit (25150-25)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 40 ppb (K-7540)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 1 ppm (K-7501)
CHEMets Colorimetric Dissolved Oxygen Test Kit, 1 to 12 ppm (K-7512).
More Details
Scuba Fill Tank Water Bath
Buy Scuba Fill Tank Water Bath Items Including:
Dual-Chamber Poly Water Bath, 5.5-liter capacity each chamber, ambient (YO-39066-50)
Plexiglas tank, 8-liter capacity; measures 4-3/4"L x 17-7/4"W x 5-15/1
Plexiglas tank, 6-liter capacity; measures 4-3/4"L x 13-7/8"W x 5-15/1.
More Details
Ubbelohde Viscometer Capillary
Buy Ubbelohde Viscometer Capillary Items Including:
Cannon Ubbelohde viscometer, size 0C
Cannon Ubbelohde viscometer, size 0B
Cannon Ubbelohde viscometer, size 1C
Cannon Ubbelohde viscometer, size 1B
Cannon Ubbelohde viscometer, size 2C
More Details
Usa Standard Testing Sieve Fisher
Buy Usa Standard Testing Sieve Fisher Items Including:
Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, full height, No (YO-59940-01)
Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, full height, No (YO-59940-02).
More Details
In-Line Ph Electrode
Buy In-Line Ph Electrode Items Including:
Cole-Parmer In-Line pH Electrode with Tuff-Tip, 100Ω ATC Ele (YO-05995-02)
Cole-Parmer pH electrode, in-line, double-junction, 100 Ohm RTD w/tinn (YO-27001-72)
Cole-Parmer pH electrode, in-line, double-junction, no ATC 10-ft cable (YO-27001-90).
More Details
Orion Triode
Buy Orion Triode Items Including:
Thermo Scientific Orion pH/ATC Triode, epoxy body, sealed, single-junc (9107WMMD)
Thermo Scientific Orion ROSS Ultra pH/ATC Triode, epoxy body, refillab (8157BNUMD)
Thermo Scientific Orion ROSS Ultra pH/ATC Triode, epoxy body, refillab (8157UWMMD).
More Details
1 Ml Pipette
Buy 1 Ml Pipette Items Including:
Cole-Parmer Disposable Pipette Tips, 3-9/16" L, 200 µL - 1 mL, 600/
Kimble Disposable Serological Non-sterile Pipettes, Glass, 1 mL, To De
Kimble Disposable Serological Sterile Plugged Pipettes, Glass, 1 mL, T
Pyrex 7077 Pipette, 1 ML, 800/CS.
More Details
Battery Operated Full Face Respirator
Buy Battery Operated Full Face Respirator Items Including:
North Full-Face Respirator, Thermoplastic elastomer
3M 6000-series Full-face respirators, Small
Survivair Opti-Fit™ Full-Face Respirator, Small
3M FF-400 Series Full-Facepiece Respirators, Small
3M FF-400 Series Full-Facepiece Respirators, Medium.
More Details
Duran Glass Laboratory Bottles
Buy Duran Glass Laboratory Bottles Items Including:
Clear glass dropping bottle, 100 mL
Pack of 6
(W211735)
Clear glass dropping bottle, 50 mL
Pack of 6
More Details
Autoclave Carboy W Spigot 20 Liter
Buy Autoclave Carboy W Spigot 20 Liter Items Including:
Economical heavy duty carboy with spigot, 20 liter
Thermo Scientific Nalgene polycarbonate carboy with spigot, 20 L
Economical heavy duty carboy with spigot, 5 liter
Thermo Scientific Nalgene low-density polyethylene carboy with spigot (2318-0050).
More Details
Nalgene 25 Ml Volumetric Flask
Buy Nalgene 25 Ml Volumetric Flask Items Including:
Thermo Scientific Nalgene Class B polypropylene volumetric flask, 50 m
Thermo Scientific Nalgene Class B polypropylene volumetric flask, 1000
PYREX Brand 5640 Volumetric Flask; 25 mL, case of 12
PYREX Brand 5580 Volumetric Flask; 25 mL, case of 12.
More Details
Chemical Resistant Gloves Category A
Buy Chemical Resistant Gloves Category A Items Including:
Best Nitri-Solve Knit, Solvent Resistant Gloves, size Medium, 14" long
Ansell® Chemical-Resistant PVA Gloves, 12"L, Size 9, 12 pair/pk
Best Viton Gloves, small, 12-mil thickness, 1 pair
Best Viton Gloves, medium, 12-mil thickness, 1 pair.
More Details
Ph Tester 30
Buy Ph Tester 30 Items Including:
Oakton Waterproof pHTestr 30 Pocket pH Tester
Horiba Laqua Tester, Calcium
accumet® AB 30 benchtop conductivity meter kit
accumet® XL 30 benchtop conductivity meter only
YSI 30 handheld conductivity meter with cell and 10-ft cable
More Details
Handheld Temperature Sensor
Buy Handheld Temperature Sensor Items Including:
Fluke 68 Infrared Thermometer, Advanced high-temperature model (FLUKE-68)
Non-evasive moisture meter with high frequency pinless sensor
Fluke® 62 Compact Infrared Thermometer (10:1)
C570 Pouch and carrying kit, Includes padded pouch and belt clip.
More Details
Plastic Lab Containers for Flammable Liquids
Buy Plastic Lab Containers for Flammable Liquids Items Including:
Safety Sign, Danger - Flammable Liquids, 10" X 14", Plastic
Cole-Parmer Disposable Lab Container, Polypropylene Copolymer (PPCO), (YO-06727-00)
Cole-Parmer Disposable Lab Container, Polypropylene Copolymer (PPCO), (YO-06727-10).
More Details
Large Od Tubing Electric Pinch Valve
Buy Large Od Tubing Electric Pinch Valve Items Including:
Manual Pinch Valve, 5/32" to 1/4" tube OD (PV-2)
Manual Pinch Valve, 5/16" to 3/8" tube OD (PV-3)
Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 10 mm Tube OD, (PK-0802-NC-3 12V N CLOSED).
More Details
Aalborg Flow, Level & Valves
Buy Aalborg Flow, Level & Valves Items Including:
Aalborg Stepping Motor Proportioning Valve (aluminum), 200 sL/min (air
Aalborg Stepping Motor Proportioning Valve (stainless steel), 200 sL/m
Aalborg Stepping Motor Proportioning Valve (aluminum), 500 sL/min (air.
More Details
Laboratory Screens
Buy Laboratory Screens Items Including:
Spectra/Mesh Screen Sheets, Polypropylene, 500 µm, 3/pk
Spectra/Mesh Screen Sheets, Polypropylene, 350 µm, 3/pk
Spectra/Mesh Screen Sheets, Polypropylene; pore size, 210µ; 30 x 30
Spectra/Mesh Screen Sheets, Polypropylene, 30 x 30 cm, 149 µm, 3/pk.
More Details
Digi-Sense Thermocouple Probes
Buy Digi-Sense Thermocouple Probes Items Including:
Type J Industrial Direct Insert Ungrounded Thermocouple Probe 12"L (700-J-F-12-W4-316-AL-1/2)
Type K Industrial Direct Insert Ungrounded Thermocouple Probe 12"L (700-K-F-12-W4-316-AL-1/2)
Carrying case for Digi-Sense meter (1316-1).
More Details
Specific Gravity Scales
Buy Specific Gravity Scales Items Including:
H-B Instrument Dual-Scale Hydrometer, 0.700 to 2.000 Specific Gravity,
Durac Baume & Specific Gravity Hydrometer, 1.800/2.000 Sp Gr, 0.005 Di
H-B Instrument Dual-Scale Hydrometer, 1.000 to 2.000 Specific Gravity, (50270).
More Details
Nist Traceable Mercury Thermometer Astm
Buy Nist Traceable Mercury Thermometer Astm Items Including:
NIST-Traceable Calibration; Thermometer Probe used with IR Thermometer (YO-17004-10)
Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (062CBBLSFC)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with genera (1340-5417).
More Details
Cryogenic Gloves
Buy Cryogenic Gloves Items Including:
Water-resistant Cryo-Glove gloves, 12"L, medium, 1 pair
Water-resistant Cryo-Glove gloves, 12"L, large, 1 pair
Cryo-Industrial gloves, mid-arm style, medium, 15" length
Cryo-Industrial gloves, mid-arm style, large, 15" length.
More Details
Sterile Sampling Bags
Buy Sterile Sampling Bags Items Including:
Whirl-Pak sterile sampling bag, 18 oz
Whirl-Pak sterile sampling bag, 24 oz
Whirl-pak Sterile Sampling Bag, 69 oz 500/bx
Whirl-Pak sterile sampling bag with white labeling area, 2 oz
Whirl-Pak sterile sampling bag with white labeling area, 18 oz.
More Details
Glass Chromatography Columns
Buy Glass Chromatography Columns Items Including:
Corning Standard Glass Chromatography Column, 10.5 ID x 150 L
Corning Standard Glass Chromatography Column, 10.5 ID x 250 L
Corning Standard Glass Chromatography Column, 19.0 ID x 250 L
Corning Standard Glass Chromatography Column, 19.0 ID x 500 L.
More Details
Natural Gas Pressure Gauge
Buy Natural Gas Pressure Gauge Items Including:
Special-Duty Air-Operated Natural Gas Powered Double-Diaphragm Pump
Special-Duty Air-Operated Natural Gas Powered Double-Diaphragm Pump, 1 (G1FB1ABTXNSX00)
Special-Duty Air-Operated Natural Gas Powered Double-Diaphragm Pump, 1 (G15B1ABTXNSX00).
More Details
Stainless Steel to Plastic Fittings
Buy Stainless Steel to Plastic Fittings Items Including:
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41).
More Details
70618
results
<<<Previous
13
14
15
16
17
18
19
20
21
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Air Products Regulators to Stainless Steel to Plastic Fittings
>
31125329-s-toc_2 31125329stoc2