Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 18
Laboratory Hot Plate & Slides
to
Domnick Hunter Filters
Image
Product Detail
More Detail
Laboratory Hot Plate & Slides
Buy Laboratory Hot Plate & Slides Items Including:
Lab
Companion® Stirring Hot Plate, Ceramic Top, 240V (BDH571065K)
Lab
Companion® Stirring Hot Plate, Ceramic top, 120V (BDH571066U)
IKA RH Basic Stirring Hot Plate, 115V
More Details
Kynar Tubing
Buy Kynar Tubing Items Including:
Kynar tubing, 11/64"ID x 1/4"OD, 25 ft/pack
Kynar® tube set for 07075-series motors, 40"L (16520-3400)
Tubing Adapter; Kynar, 1/2" mini to 1/4" NPT (M)
Tubing Adapter; Kynar, 1/2" mini to 3/8" NPT (M).
More Details
Masterflex Easy Load
Buy Masterflex Easy Load Items Including:
Replacement rotor assembly for Masterflex L/S Easy-Load II pump heads (YO-77200-58)
Tubing retainer kit for I/P Easy-Load 77601-series polysulfone pump he
Masterflex L/S Easy-Load II pump head for precision tubing, fixed occl (YO-77200-50).
More Details
Norprene High Pressure Peristaltic Pump
Buy Norprene High Pressure Peristaltic Pump Items Including:
Pump, Cole-Parmer Deluxe Peristaltic Metering Pump, Max flow of 17.4 G (YO-74200-56)
Cole-Parmer Norprene Replacement Roller set for 74200 series of pumps, (A3-SNF-R)
Cole-Parmer Norprene Replacement Roller set for 74200 series of pumps, (A3-SNH-R).
More Details
Nist Thermometers
Buy Nist Thermometers Items Including:
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with genera (1340-5417)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with penetr (1340-5418)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with handle (1340-5416).
More Details
Oakton Instruments
Buy Oakton Instruments Items Including:
Oakton Waterproof pH 600 Meter Only
Oakton Waterproof pH/DO 300 Meter Only
Replacement plastic storage box for Oakton Testrs
Conductivity/Temp probe, K=10.0 for Oakton 6 and 6+ series meters
Oakton Temp-14 Thermistor Thermometer w/boot (CPTEMP14).
More Details
Parker Ball Valve
Buy Parker Ball Valve Items Including:
Parker two-way straight ball valve, stainless steel, 1/8" compression
Parker two-way straight ball valve, stainless steel, 1/4" compression
Parker two-way straight ball valve, stainless steel, 3/8" compression.
More Details
0.2 Micron Capsule Filter with Npt
Buy 0.2 Micron Capsule Filter with Npt Items Including:
Whatman™ Disposable Capsule Filters, PTFE Membrane; pore size, 1.0 (YO-29700-02)
Whatman™ Disposable Capsule Filters, PTFE Membrane; pore size, 0.2 (YO-29700-00)
Filter Cartridge, Pleated All-Polypropylene, 0.2 micron absolute-rated (HC-PP-40-0.2).
More Details
Npt X 3 8 Compression Fitting
Buy Npt X 3 8 Compression Fitting Items Including:
Compression pipe adapter, 3/8" NPT x 3/8", Brass, each (YO-31412-36)
Compression fitting; probe diameter, 3/8"; 316SS; 3/8"NPT (M) (6-6 FH4BZ-SS-C)
Pharmafluor Compression To Male NPT Thread Adapter, PFA, 1/4 X 3/8 (YO-31001-35).
More Details
Low Flow Switch
Buy Low Flow Switch Items Including:
Compact low-flow switch for gases; SPST normally open, 25 cfh/60 cfh (YO-32778-30)
Compact low-flow switch for gases; SPST normally open, 1 cfm/2.6 cfm (YO-32778-32)
Adjustable Standard Low-Flow Switch for Liquids and Gases, 3.0/500 cc/ (125-BBPNC).
More Details
Ph66 Ss Ph Probe
Buy Ph66 Ss Ph Probe Items Including:
ISFET pH probe, general purpose (PHW77-SS)
Repl SS probe for IQ240 benchtop meter
SS probe with heavy-duty handle for IQ150 handheld 59540-22
SS piercing tip, micro probe for IQ240 benchtop meter
General purpose ISFET probe, waterproof (PHW17-SS).
More Details
Plastic Hydrometer
Buy Plastic Hydrometer Items Including:
Thermo Scientific ERTCO® Plastic Hydrometer, 0 to 26.5 Percent Sal (2511-02PL)
Durac Baume Polycarbonate Hydrometer, 19 to 31, 0.2 Div, 255mm L, Indi
Durac Brix Sugar Scale Polycarbonate Hydrometer, 0 to 35, 0.5 Div, 300.
More Details
Laboratory Stir Plates
Buy Laboratory Stir Plates Items Including:
Lab
Companion® Stirring Hot Plate, Ceramic Top, 240V (BDH571065K)
IKA RH Digital Stirring Hot Plate, 115V
PTFE-coated lab spatula, with 1/2"L spooned end and 1 1/2"L flat round (D1069292)
IKA ETS-D6 contact thermometer with pH (3378100).
More Details
Tris Compatible Ph Electrode
Buy Tris Compatible Ph Electrode Items Including:
Accumet pH Electrode, Combination, Microprobe, Refillable, Glass Body,
Accumet pH Electrode, Combination, General Purpose, Long-neck, Gel-fil
Honeywell Durafet III, non-glass pH electrode, 1.5 inch Tri-Clamp flan.
More Details
100 Ml Centrifuge Tube Rack
Buy 100 Ml Centrifuge Tube Rack Items Including:
Centrifuge tube rack for 15-mL centrifuge tubes (YO-06696-02)
Test tube racks for 1.5 mL tube size; PP (YO-06703-40)
Costar Spin-X Centrifuge Tube Filters, Spinfilter Ca 0.22um 100/cs
Centrifuge Tube Ice Rack (holds 28 X 50 mL Tubes) (F18905-0050).
More Details
Ph Probe Flow Cells
Buy Ph Probe Flow Cells Items Including:
Cole-Parmer Flow Cell for Ultrapure Water, 316 SS
Flow cell, Schedule 80 CPVC, 3/4" x 3/4" FNPT
Conductivity probe, 2-cell, K = 0.1, epoxy body/platinum electrode, 2-
Conductivity probe, 2-cell, K = 1.0, Ultem body/stainless steel electr.
More Details
Female Luer Lock Connector
Buy Female Luer Lock Connector Items Including:
Adapter, PP, 1/2" mini to female luer lock (YO-31200-50)
Adapter, Kynar, 1/2" mini to 1/2" to female luer lock
Female luer lock x 1/8" NPT, 316 SS, each (YO-31507-46)
Female luer lock x 1/4" NPT, 316 SS, each (YO-31507-48).
More Details
Cryogenic Storage Racks
Buy Cryogenic Storage Racks Items Including:
Polypropylene cryogenic vial rack, holds 50 round bottom vials (985800)
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 81 ( (5026-0909)
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 81 ( (5027-0909).
More Details
Latex Free Plastic Centrifuge Tube
Buy Latex Free Plastic Centrifuge Tube Items Including:
15 mL conical bottom disposable plastic centrifuge tubes, screw cap/ri (3219P)
15 mL conical bottom disposable plastic centrifuge tubes, screw cap/ri (3215P)
15 mL conical bottom disposable plastic centrifuge tubes, screw cap/ri (3220P).
More Details
Tubing Cross Clamp
Buy Tubing Cross Clamp Items Including:
Sanitary Clamp Cross, 316L SS, 1" (YO-30560-42)
SANITARY CLAMP CROSS, 316L SS, 1-1/2" (YO-30560-43)
SANITARY CLAMP CROSS, 316L SS, 2" (YO-30560-44)
Aluminum cross, NW 16 (C10512412)
Cross Fittings, 100 ISO-K, aluminum (C100-09-211).
More Details
Watch Glass on Base Chemical Supplies
Buy Watch Glass on Base Chemical Supplies Items Including:
Pyrex Brand 9985 watch glass; 100 mm, pack of 12
Pyrex Brand 9985 watch glass; 125 mm, pack of 12
Pyrex Brand 9985 watch glass; 150 mm, pack of 12
Pyrex Brand 9985 watch glass; 65 mm, case of 144
Pyrex Brand 9985 watch glass; 100 mm, case of 144.
More Details
Dry Glass Bead Sterilizer Suppliers
Buy Dry Glass Bead Sterilizer Suppliers Items Including:
Dry Glass Bead Sterilizer, 3" D x 1-3/4" dia sterilizing well; 110 VAC (YO-10779-00)
Dry Glass Bead Sterilizer, 3" D x 1-3/4" dia sterilizing well; 220 VAC (YO-10779-05).
More Details
Parker Porter Gas Mass Flow Controllers
Buy Parker Porter Gas Mass Flow Controllers Items Including:
Panel Mount Mass Flow Controller 0.5 SLPM; 1/8" NPT(F); 24 VDC
Panel Mount Mass Flow Controller 2.0 SLPM; 1/8" NPT(F); 24 VDC
Panel Mount Mass Flow Controller 5 SLPM; 1/8" NPT(F); 24 VDC.
More Details
Oakton Hygrothermograph
Buy Oakton Hygrothermograph Items Including:
Two-Speed, Spring-Wound Hygrothermograph (7215-00)
Long-Cycle Hygrothermograph (7012-00)
Optional axial fan, for more uniform sample measurments in fluctuating (7210-30)
Chart paper, for long cycle, -20 to 50°C, 1-month, 2-hour incremen (7012-70).
More Details
Tubing Ptfe 1 16 Od
Buy Tubing Ptfe 1 16 Od Items Including:
Compress to male threaded adapter, Pharmafluor PTFE, 1/16" OD x 1/8" N
Replacement tubing for flow adapters, FEP PTFE, 1⁄16" OD x 10 ft (YO-34696-74)
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02).
More Details
Syringe Filters Polypropylene Housing 25mm
Buy Syringe Filters Polypropylene Housing 25mm Items Including:
Cole-Parmer HPLC Syringe Filters, Non Sterile, Polypropylene (PP), 0.4 (YO-32816-94)
Cole-Parmer HPLC Syringe Filters, Non Sterile, Polypropylene (PP), 0.2 (YO-32816-96)
Whatman Anotop syringe filter, 25mm, 0.02µm, sterile, 50/pk (6809-2102).
More Details
Bacterialogical Water Sampling Bottles
Buy Bacterialogical Water Sampling Bottles Items Including:
Thermo Scientific Nalgene Storm Water Sampler Bottle, HDPE for Inorgan
Thermo Scientific Nalgene Storm Water Sampler Bottle, Amber Glass for
Cap for Oakton sample bottles, pack of 10
Graduated Sampler Bottles, 100 mL, PP, Case of 100 (202445-0100).
More Details
Millipor 45 Micron Filter Paper
Buy Millipor 45 Micron Filter Paper Items Including:
Millipore Syringe Filters, MF-Millipore MCE, .45µm 50pk
Millipore Syringe Filters, Millipore LCR PTFE, .45µm 50pk
Millipore Syringe Filters, Durapore PVDF, .45µm 50pk (YO-81053-12)
Millipore Syringe Filters, Durapore PVDF, .45µm 50pk (YO-81053-24).
More Details
Advantec Mixed Cellulose Ester Membrane Filters
Buy Advantec Mixed Cellulose Ester Membrane Filters Items Including:
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.6 (YO-06644-06)
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.4 (YO-06644-08).
More Details
Domnick Hunter Filters
Buy Domnick Hunter Filters Items Including:
Parker Domnick Hunter Compressed Air Activated Carbon Filter, 21 SCFM,
Parker Domnick Hunter Compressed Air Activated Carbon Filter Element, (YO-29587-21)
Parker Domnick Hunter Compressed Air Activated Carbon Filter Element, (YO-29587-23).
More Details
70618
results
<<<Previous
14
15
16
17
18
19
20
21
22
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Laboratory Hot Plate & Slides to Domnick Hunter Filters
>
31125329-s-toc_2 31125329stoc2