Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 19

10 Ml Graduated Cylinder   to   Whatman 50 Micron Filter Paper

Image
  Product Detail
More Detail

10 Ml Graduated Cylinder

Buy 10 Ml Graduated Cylinder Items Including:
  • Pyrex Brand 3024 cylinder; 10 mL, pack of 1
  • Pyrex Brand 3024 cylinder; 10 mL, case of 24
  • Pyrex Brand 2982 cylinder; 10 mL, case of 24
  • Cylinder, Pyrex Brand 3025, graduated, 10 mL
  • Pyrex Brand 3042 cylinder; 10 mL, pack of 1 (3042-10) (YO-34547-01).

1 Liter Volumetric Flask

Buy 1 Liter Volumetric Flask Items Including:
  • Volumetric Flask, PMP, 25mL, 1/pk (YO-06110-32)
  • Volumetric Flask, PMP, 100mL, 1/pk (YO-06110-36)
  • Volumetric Flask, PMP, 250mL, 1/pk (YO-06110-38)
  • Volumetric Flask, PMP, 500mL, 1/pk (YO-06110-40)
  • PYREX Brand 5640 Volumetric Flask; 1 mL, case of 12.

Fluke 51 Thermocouple Meter

Buy Fluke 51 Thermocouple Meter Items Including:
  • Fluke 50-Series II (Model 51) Thermocouple Thermometer with Single In (FLUKE-51-2 60HZ)
  • Fluke 50-Series II (Model 52) Thermocouple Thermometer with Dual Inpu (FLUKE-52-2 60HZ)
  • Fluke 50-Series II (Model 54) Thermocouple Thermometer Dual Input wit.

3 Way Stopcocks Pharmaseal K75

Buy 3 Way Stopcocks Pharmaseal K75 Items Including:
  • PTFE 2-Way Stopcock, 3/8" Orifice, 3/8" NPT(F) connections
  • High Pressure Stopcock, 1050 psi max, 3-way, 10 pack (YO-30526-34)
  • Large-bore 3-way, male-lock, stopcocks, 10/pack (YO-30600-23)
  • 3-way male lock stopcock, 100/pack (YO-30600-25).

Absolute Pressure Gauge Mmhg

Buy Absolute Pressure Gauge Mmhg Items Including:
  • Cole-Parmer Battery-powered Digital Gauge, 760 to 0 Torr absolute; 1/
  • Cole-Parmer Battery-powered Digital Gauge, 760.0 to 0 Torr absolute w/
  • Cole-Parmer Loop-powered Digital Gauge, 760 to 0 Torr absolute; 1/4".

Eppendorf Pipette Tips 20 200 Ul

Buy Eppendorf Pipette Tips 20 200 Ul Items Including:
  • 0.1 to 20µL epTIPS pipette tips, Starter set (022491415)
  • 20 to 300µL epTIPS pipette tips, Starter set (022491440)
  • 0.1 to 20µL epTIPS pipette tips, bulk pack of 1000 (022492012)
  • 20 to 300µL epTIPS pipette tips, bulk pack of 1000 (022492047).

Barnstead Laboratory Equipment

Buy Barnstead Laboratory Equipment Items Including:
  • Nanopure UV Systems (RO/Distilled Feed) Consumables Bundle (D502142)
  • Nanopure UV Systems (Deionized Feed) Consumables Bundle (D502141)
  • Nanopure Life Science Systems (RO/Distilled Feed) Consumables Bundle (D502144).

Evaporating Dishes

Buy Evaporating Dishes Items Including:
  • Dish, evaporating, porcelain, glazed inside and partly outside, standa (60204)
  • Dish, evaporating, porcelain, glazed inside and partly outside, standa (60205)
  • Dish, evaporating, porcelain, glazed inside and partly outside, standa (60206).

Compound Pressure Gauge 30 0 600

Buy Compound Pressure Gauge 30 0 600 Items Including:
  • Low-Cost Utility Pressure Gauge, 30" Hg to 30 psi (KCCP7211-2.5-4-30/30)
  • Additel Digital Pressure Gauge, 0 to 600 psi, 1/4" NPT(M) Connection
  • SPAN Pressure Gauge, 0 to 600 psi (YO-68081-50)
  • SPAN Pressure Gauge, 0 to 600 psi (YO-68081-20).

Replacement Electrode Phtestr 1 2

Buy Replacement Electrode Phtestr 1 2 Items Including:
  • Free Chlorine Replacement Electrode
  • GLI replacement compression fitting, 1/2", kynar (1000F1236-111)
  • Ultrasonic Cell Disruptor Replacement Tips, 1/2" dia
  • Mounting Brackets for 3/8" and 1/2" NPT(F) elliptic valves, 2/set (UMB-2).

Gf Signet Flow Monitors and Controllers

Buy Gf Signet Flow Monitors and Controllers Items Including:
  • Digital Flow Transmitter and Display, +GF+ Signet Loop Powered (7-30 V
  • Multi-parameter controller; 110/220 VAC, 50/60 Hz (3-8900+401-1+402-1)
  • Batch Controller, 12-24 VAC/dc (3-5600).

National Weather Service Rain Gauge

Buy National Weather Service Rain Gauge Items Including:
  • Rainwise Dual Display Rain Gauge, 30ft cable
  • Rainwise Wireless Dual Display Rain Gauge
  • Cole-ParmerClear Rain Gauges, 0.01 to 11.0"
  • Cole-ParmerClear Rain Gauges, 0 to 25.2 cm
  • TR-525I : Rain Gauge, Tipping Bucket 6" Collector.

Luer Lock Adapter

Buy Luer Lock Adapter Items Including:
  • Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
  • Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
  • Adapter, Kynar, 1/2" mini to 1/2" to female luer lock
  • 316 SS fittings; male luer lock plug (YO-31507-23).

Water Conductivity Standards

Buy Water Conductivity Standards Items Including:
  • Cole-Parmer conductivity standard, 1 µS, 16 ounces
  • Cole-Parmer One-Shot™ conductivity standards, 5 µS, 100 mL vials
  • Cole-Parmer One-Shot™ conductivity standards, 10,000 µS, 100 mL
  • Cole-Parmer One-Shot™ conductivity standard assortment pack, 100 mL.

Nalgene Tubing

Buy Nalgene Tubing Items Including:
  • Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
  • Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o
  • Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, y.

Pvc 3 Way Ball Valves

Buy Pvc 3 Way Ball Valves Items Including:
  • Hayward electrically Actuated Ball Valve, 3-way, 1/2" PVC (HCTN1050STV/EAU-28)
  • Hayward electrically Actuated Ball Valve, 3-way, 1" PVC (HCTN1100STV/EAU-28)
  • Hayward electrically Actuated Ball Valve, 3-way, 2" PVC (HCTN1200STV/EAU-28).

High Shear Mixer Blade

Buy High Shear Mixer Blade Items Including:
  • Lightnin® 316 SS high-shear radial flow impeller (Lightnin® R- (YO-04560-30)
  • Lightnin® 316 SS high-shear radial flow impeller (Lightnin® R- (YO-04560-56)
  • Three-stage high shear dispersing mixing head for 04300-01 Magic Lab ( (R078352).

Reference Electrode Filling Solution

Buy Reference Electrode Filling Solution Items Including:
  • ROSS electrode fill solution, 3 M KCl
  • Case of five 60-mL bottles (810007)
  • Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
  • Filling solutions: use with Cl- electrodes (900017)
  • Filling solution f/ammonia ISE electrode (951202).

Ashcroft Vacuum Gauges

Buy Ashcroft Vacuum Gauges Items Including:
  • Ashcroft Test Gauge, 0.25% Accuracy, Bronze Bourdon Tube, 4 1/2" Dial, (10824.5) (YO-68030-01)
  • Ashcroft Test Gauge, 0.25% Accuracy, Bronze Bourdon Tube, 0 to 15 psi
  • Ashcroft Test Gauge, 0.5% Accuracy, 316 SS Bourdon Tube, 3" Dial, 30 t.

Minipuls 2 Pump Gilson Tubing

Buy Minipuls 2 Pump Gilson Tubing Items Including:
  • Ismatec™ tubing-bed cartridge set, 2 channels, nominal tubing (YO-07611-10)
  • Masterflex L/S Easy-Load ATEX Zone 2 pump head for precision tubing
  • Masterflex L/S Easy-Load ATEX Zone 2 pump head for high-performance pr.

Corning Bottletop/Filter Units

Buy Corning Bottletop/Filter Units Items Including:
  • Corning Disposable Bottletop Filter; 150ml 0.45µm
  • Corning Disposable Bottletop Filter; 150-mL volume; PES, membrane; 48/
  • Corning Disposable Bottletop Filter; 150-mL volume; CA, membrane; 48/p
  • 150 mL tube top vacuum filters, 0.22 µm, 12/cs (430320).

Glove Bags

Buy Glove Bags Items Including:
  • Glove Bag™ Inflatable Glove Chamber with Two Equipment Sleeves, 12" (YO-04408-40)
  • Glove Bag™ Inflatable Glove Chamber, 33 1/2" Equipment Sleeve (YO-04408-58)
  • Glove Bag™ Inflatable Glove Chamber, 33 1/2" Equipment Sleeve (YO-04408-60).

Hand Operated High Pressure Water Pump

Buy Hand Operated High Pressure Water Pump Items Including:
  • Hand-operated vacuum/pressure pump, 15 mL/stroke, zinc alloy body
  • Hand-operated vacuum pump (without gauge), 32 mL/stroke, PVC body
  • Low pressure test pump, generates -12 psi/-0.85 bar or pressures to 30
  • Repair kit for vacuum/pressure pump 79301-11.

Ph Standard Solutions

Buy Ph Standard Solutions Items Including:
  • Standardizing solution: 0.1 M NaNO3 (920706)
  • Standardizing solution: 0.1 M CaCl2 (922006)
  • Standardizing solution: NaF as F, 100 ppm (940907)
  • Standardizing solution: NaCl as Na, 100 ppm (941107)
  • Standardizing solution: NaCl as Cl, 100 ppm (941707).

Tubing Formulation Tygon E-Food (B-44-4x)

Buy Tubing Formulation Tygon E-Food (B-44-4x) Items Including:
  • Masterflex Tygon E-Food (B-44-4X) tubing, B/T 87, 10 ft.
  • Masterflex Tygon E-Food (B-44-4X) tubing, B/T 91, 10 ft.
  • Masterflex Tygon E-Food (B-44-4X) tubing, I/P 88 tubing, 50' ft.
  • Masterflex Tygon E-Food (B-44-4X) tubing, I/P 89 tubing, 50' ft..

Individually Wrapped Sterile Pipette Tips

Buy Individually Wrapped Sterile Pipette Tips Items Including:
  • Thermo Scientific Finntip 10 mL Sterile Pipette Tips, 2 to 10 mL, 5x24
  • Thermo Scientific Finntip ® 300 Sterile Pipette Tips, 5-300 µL, 10 (9401253)
  • Reagent Reservoir, 25 mL, 50/case, Individually wrapped, Sterile, Pyro.

Stainless Steel Beakers

Buy Stainless Steel Beakers Items Including:
  • Stainless-steel Griffin-style beaker with easy-pour rim, 125 mL (YO-07205-30)
  • Immersion heater, beaker, 2.75" dia
  • heated area, 550 watts, 115 VAC (297-2)
  • Immersion heater, beaker, 2.75" dia
  • heated area, 550 watts, 230 VAC (297-2).

Plastic Pipette

Buy Plastic Pipette Items Including:
  • Transfer Pipette, 1.5ml, Graduated, 500/pk (YO-06226-37)
  • Cole-Parmer sterile Pipette, bulk wrapped, plastic green TD ring, 25mL
  • Cole-Parmer sterile Pipette, bulk wrapped, plastic purple TD ring, 50m
  • Disposable Droppers, LDPE, 5 mL, 100/pk (262185).

Glass Sample Bottles

Buy Glass Sample Bottles Items Including:
  • I-CHEM pre-cleaned sample jars with certificate of analysis, 1250 mL, (341-1250)
  • Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 30 mL, 20 mm Ne
  • Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 60 mL, 20 mm Ne.

Whatman 50 Micron Filter Paper

Buy Whatman 50 Micron Filter Paper Items Including:
  • Whatman Glass Microfiber Filter Papers Water-Pollution test papers; 4
  • (YO-06648-70)
  • Whatman Glass Microfiber Filter Papers Water-Pollution test papers; 4
  • (YO-06648-71).



10 Ml Graduated Cylinder to Whatman 50 Micron Filter Paper

>