Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 20
Taco 24 Volt 2 Way Valve
to
304 Stainless Steel Tubing Suppliers
Image
Product Detail
More Detail
Taco 24 Volt 2 Way Valve
Buy Taco 24 Volt 2 Way Valve Items Including:
Ball Valve, 2-way, 1/4" NPT(F) x 1/4" NPT(M), PVC with EPDM seals (YO-01377-24)
Ball Valve, 2-way, 3/8" barb, PVC with EPDM seals (YO-01378-24)
Particle Tolerant Solenoid Valve, two-way, normally closed, 24 VDC, 2
(MTV-2K.25U-G-24V).
More Details
Stainless Steel Surgical Tweezers
Buy Stainless Steel Surgical Tweezers Items Including:
Tweezers, stainless steel, straight, strong precision tips, 5"L (18.013USA)
Tweezers, stainless steel, straight, serrated grips, thick, flat, stro (18.037USA)
Tweezers, stainless steel, straight, ultra fine sharp tips, 4-1/4"L (18.056USA).
More Details
8 Inch Sieve
Buy 8 Inch Sieve Items Including:
Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8” OD, full height, No
Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, half height, No (YO-59941-00)
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-50).
More Details
Chemical Spatula
Buy Chemical Spatula Items Including:
SMART SPATULA™ Disposable Spatula Opaque 210 mm (17221)
SMART SPATULA™ Disposable Anti static Micro Spatula, Opaque 140 mm (17231)
SMART SPATULA™ Disposable Macro Spatula, Green 310 mm (17241)
SMART SPATULA™ Disposable Spatula, blue 210 mm (17211).
More Details
Wastewater Sampling Equipment
Buy Wastewater Sampling Equipment Items Including:
Global Water WS755 Improved Wastewater/Stormwater Sampler (YO-99243-15).
More Details
50 Ml Petri Dish
Buy 50 Ml Petri Dish Items Including:
Chemware Petri Dish PFA, 100 mL
Petri dish with pads, 50x11 mm, 100/cs (YO-14005-20)
Petri dish without pads, 50x11 mm, 100/cs
Petri dish without pads, 50x11 mm, 500/cs
Petri dish with pads, 50x11 mm, 500/cs
More Details
Hot Plate with Surface Temperature Control
Buy Hot Plate with Surface Temperature Control Items Including:
RET basic IKAMAG Stirring Hot Plate 115V (3622001)
RET basic IKAMAG stirring hot plate 230V (3622000)
TSI™ VelociCalc Surface Temperature Probe
Surface-Mount Refrigeration Alarm; temperature range; -10 - 80F, 110V (TA-2F-110).
More Details
Zero Oxygen Standard Solution
Buy Zero Oxygen Standard Solution Items Including:
OptiSphere™ 0.2 NTU Turbidity Standard, 250-mL
OptiSphere™ 0.5 NTU Turbidity Standard, 250-mL
OptiSphere™ 200 NTU Turbidity Standard, 250-mL
OptiSphere™ 800 NTU Turbidity Standard, 250-mL
OptiSphere™ 1000 NTU Turbidity Standard, 250-mL.
More Details
Ph Calibration Standards
Buy Ph Calibration Standards Items Including:
Conductivity calibration standard, 1000 µmhos (3161)
MYRON L conductivity calibration standard, 23.8 µS
MYRON L conductivity calibration standard, 46.7 µS
MYRON L conductivity calibration standard, 2060 µS.
More Details
Oakton Ecotestr Salt Pocket Salinity Tester
Buy Oakton Ecotestr Salt Pocket Salinity Tester Items Including:
Oakton SaltTestr pocket salinity tester (YO-35661-52)
Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
Oakton EcoTestr TDS Low pocket TDS tester
Oakton EcoTestr EC Low pocket conductivity tester
Oakton EcoTestr EC High pocket conductivity tester.
More Details
Tygon 2275 Ib High-Purity Reinforced Tubing
Buy Tygon 2275 Ib High-Purity Reinforced Tubing Items Including:
Tygon High Purity Reinforced Tubing, 3/4"ID x 1-1/16" OD, 50 ft/pk
Tygon High Purity Reinforced Tubing, 1/4"ID x 1/2"OD, 50 ft/pk
Tygon High Purity Reinforced Tubing, 3/8"ID x 5/8"OD, 50 ft/pk.
More Details
Nist Traceable Sieve Calibration Glass Beads
Buy Nist Traceable Sieve Calibration Glass Beads Items Including:
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-50)
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-51).
More Details
Brady Labxpert Label Printers and Labels
Buy Brady Labxpert Label Printers and Labels Items Including:
Label, For Labxpert, For Tube/vials (YO-65544-02)
Label, For Labxpert, For Tube/vials (YO-65544-04)
Label, For Labxpert, For Tubes/vials (YO-65544-06)
Label, F/labxpert, For Vial Tops (YO-65544-16)
Label, F/labxpert, Tube/vial Combo (YO-65544-20).
More Details
Propane Gas Brass Needle Valve
Buy Propane Gas Brass Needle Valve Items Including:
Parker needle valve; 1/4" compression connectors, brass
Needle Valve, Brass, 600 Max psi
Bunsen burner, adjustable with threaded needle valve, natural gas (H-5885)
Bunsen burner, adjustable with threaded needle valve, artificial gas (H-5880).
More Details
Whirl Pak Bags
Buy Whirl Pak Bags Items Including:
Whirl-Pak sterile sampling bag, 18 oz
Whirl-Pak sterile sampling bag, 24 oz
Whirl-pak Sterile Sampling Bag, 69 oz 500/bx
Whirl-Pak sterile sampling bag with white labeling area, 2 oz
Whirl-Pak sterile sampling bag with white labeling area, 18 oz.
More Details
Ph Tester 30 Oakton
Buy Ph Tester 30 Oakton Items Including:
Oakton Waterproof pHTestr 30 Pocket pH Tester and calibration kit
Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
Oakton Waterproof pHTestr 10 Pocket pH Tester and calibration kit
Oakton Waterproof pHTestr 20 Pocket pH Tester and calibration kit.
More Details
Oakton Weather Products
Buy Oakton Weather Products Items Including:
Oakton Water-resistant pocket thermometer, Probe 5 in L x 0.14 in OD
Oakton Temp-16 Precision RTD Thermometer w/boot (CPTEMP16)
Oakton Temp-360 Datalogging Precision RTD Thermometer w/boot (CPTEMP360)
Oakton Thermohygrometer with Glass Thermometer.
More Details
Oakton Instruments Water Jet Aspirator Pump
Buy Oakton Instruments Water Jet Aspirator Pump Items Including:
Aspirator Pump, 18 L/min, 9.5l Bath, 115 VAC (YO-35031-00)
Aspirator Pump, 18 L/min, 9.5l Bath, 230 VAC (YO-35031-10)
Stainles Steel Aspirator for 35031-00 & -10, 1 Each (71121)
Filter Pack For 35031-00 & -10 Vacuum Pumps, 5 Pack (72200).
More Details
Kynar Rigid Tubing
Buy Kynar Rigid Tubing Items Including:
Kynar tubing, 11/64"ID x 1/4"OD, 25 ft/pack
KynarSuperflex PVDF Tubing 1/16" x 1/8" 25ft /pk
KynarSuperflex PVDF Tubing - 3/16" x 1/4" 25 ft/pk
KynarSuperflex PVDF Tubing 1/8" x 1/" 25 ft/pk.
More Details
Oakton Phtestr 2 Replacement Electrode
Buy Oakton Phtestr 2 Replacement Electrode Items Including:
Replacement electrode holder for Oakton 10 series and higher conductiv
Free Chlorine Replacement Electrode
(2) replacement membrane assemblies (35640-72) for Oakton DO probes an
Replacement pH/Con/Temp electrode w/10-ft cable for Oakton PC10 meter.
More Details
Check Valve 1 4 Flare
Buy Check Valve 1 4 Flare Items Including:
Poppet Check Valve, 1/4" NPT(F) connections (CV-1-1144)
PP Miniature Check Valve, 1/4" hose barb, Viton diaphragm
PVDF Miniature Check Valve, 1/4" hose barb, Viton diaphragm
PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV).
More Details
100 Ml Pipette
Buy 100 Ml Pipette Items Including:
CORNING Stripette Serological Pipettes, 10.0 mL, Individually Wrapped
Thermo Scientific Finntip 10 mL Pipette Tips, 2 to 10 mL, 100/bag
Kimble-Chase Volumetric Pipettes, Class A, 100 mL, case
Kimble-Chase Volumetric Pipettes, CLASS B, 100 mL, case.
More Details
250 Ml Milk Dilution Bottles
Buy 250 Ml Milk Dilution Bottles Items Including:
Corning milk dilution bottle; narrow mouth, non-graduated, 160 mL
Pac
Dilution bottle, Butterfield's phosphate buffer, 99 mL fill capacity (YO-36150-00)
Dilution bottle, phosphate buffer with magnesium chloride, 99 mL fill (YO-36150-10).
More Details
Cole-Parmer Todd Systems Inc Step-Down Transformers
Buy Cole-Parmer Todd Systems Inc Step-Down Transformers Items Including:
Todd Systems Inc SD-16 G Step-down transformer, 100 Watts
Todd Systems Inc SD-13 G Step-down transformer, 1000 Watts
Todd Systems Inc SD-14 G Step-down transformer, 1500 Watts
Todd Systems Inc SD-16-GTC Thermal Protected Step-Down Transformer, 10.
More Details
Laboratory Scissors
Buy Laboratory Scissors Items Including:
Scissor lift table, 35"L x 17-¾"W
Scissor lift table, 46"L x 19-¾"W
Precision Scissors, Straight Slim Blade (11.014)
Precision Scissors, Straight Blade (11.023)
Precision Scissors, Ceramic Straight Blade (11.802).
More Details
Digital Hygrometer Thermometer Probe Tracebale
Buy Digital Hygrometer Thermometer Probe Tracebale Items Including:
Oakton Water-resistant pocket thermometer, Probe 5 in L x 0.14 in OD
Oakton Water-resistant pocket thermometer, Probe 8 in L x 0.14 in OD
Thermocouple Probe Type K, 12" stem 1/2"NPT 1/4"diam., 316SS to 900< (DSTP1213212).
More Details
Weiss Digital Solar-Powered Industrial Pressure Gauge
Buy Weiss Digital Solar-Powered Industrial Pressure Gauge Items Including:
Pressure Gauge Vacuum-15 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socke
Pressure Gauge Vacuum-60 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socke
Pressure Gauge Vacuum-100 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Sock.
More Details
Rhodamine Wt Dye
Buy Rhodamine Wt Dye Items Including:
Flourescent FWT Red Dye Tablets, 200 tablets
Flourescent FLT Yellow/Green Dye Tablets, 200 tablets
Flourescent FLT Orange Dye Tablets, 200 tablets
Flourescent FLT Yellow/Green Dye Concentrate, Gallon
Flourescent FLT Orange Dye Concentrate, Gallon.
More Details
45 Degree Angle Pipe Fitting
Buy 45 Degree Angle Pipe Fitting Items Including:
316L Stainless Steel Asme-bpe Sanitary Clamp 45° Elbow, 20RA Mecha (YO-30606-07)
316L Stainless Steel Asme-bpe Sanitary Clamp 45° Elbow, 20RA Mecha (YO-30606-08)
316L Stainless Steel Asme-bpe Sanitary Clamp 45° Elbow, 20RA Mecha (YO-30606-09).
More Details
304 Stainless Steel Tubing Suppliers
Buy 304 Stainless Steel Tubing Suppliers Items Including:
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
304 Stainless Steel Tubing, 1/4" OD, 0.210" ID, 50ft roll (YO-34680-60)
304 Stainless Steel manifold; 2 outlets; 1/4" inlet NPT(F) (YO-31521-31).
More Details
70618
results
<<<Previous
16
17
18
19
20
21
22
23
24
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Taco 24 Volt 2 Way Valve to 304 Stainless Steel Tubing Suppliers
>
31125329-s-toc_2 31125329stoc2