Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 200

Hot Plate Stirrer Business & Industrial   to   Air Flow Meter Ppm

Image
  Product Detail
More Detail

Hot Plate Stirrer Business & Industrial

Buy Hot Plate Stirrer Business & Industrial Items Including:
  • Stirring hot plate, 4" x 5", 120 VAC
  • IKA Ceramic Stirring Hot Plate, 4" x 4", 115v (3581001)
  • IKA Ceramic Stirring Hot Plate, 10" x 10", 115v (3581401)
  • Lab
  • Companion® Stirring Hot Plate, Ceramic Top, 240V (BDH571065K).

Lmi Flow Meter 439

Buy Lmi Flow Meter 439 Items Including:
  • LMI Solenoid-Diaphragm Metering Remote-Control Pump, 2.5 GPH, 115 VAC
  • LMI Solenoid-Diaphragm Metering Remote-Control Pump, 0.42 GPH, 115 VAC
  • LMI Solenoid-Diaphragm Metering Remote Control Pump, 0.42 GPH, 230 VAC.

Water Fountain Pump Mesh Filters

Buy Water Fountain Pump Mesh Filters Items Including:
  • Filter Pack For 35031-00 & -10 Vacuum Pumps, 10 Pack (72210).

Fuji Pyz Series Temperature Controller

Buy Fuji Pyz Series Temperature Controller Items Including:
  • Temperature Controller, TC, 1/16-DIN, Relay Output, 2 alarms (DC1010CT-102-000-E)
  • Temperature Controller, TC, 1/16-DIN, Current Output, 2 alarms (DC1010CT-302-000-E)
  • Temperature Controller, TC, 1/8-DIN, Current Output, 2 alarms (DC1020CT-302-000-E).

Gast Dual Piston Vacuum Pump

Buy Gast Dual Piston Vacuum Pump Items Including:
  • Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DAA-V515-ED)
  • Vacuum/pressure diaphragm pump, single head, 1.1 cfm, 220/240VAC (DOA-P725-BN)
  • Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DOA-P704-AA).

Ptfe Filter Holders

Buy Ptfe Filter Holders Items Including:
  • Savillex PTFE 50-mm Membranes; porosity, 20-30µ; thickness, 4.8mil; (YO-06621-10)
  • Savillex PTFE 50-mm Membranes; porosity, 5-6µ; thickness, 5.6mil; 1 (YO-06621-20)
  • Savillex PTFE 50-mm Membranes; porosity, 1-2µ; thickness, 10 mil; 1 (YO-06621-30).

Wall Mount Ph Meter

Buy Wall Mount Ph Meter Items Including:
  • Eutech Instruments pH 2000 1/2-DIN Wall- Mount pH/ORP Controller
  • Wall-mount pH Controller With Recorder And Atc, One Input; 110 VAC, 50 (YO-05997-37)
  • Wall-mount pH controller with recorder, two inputs, no ATC; 110 VAC, 5 (YO-05997-80).

Pocket Ph Meter with Isfet Sensor

Buy Pocket Ph Meter with Isfet Sensor Items Including:
  • Oakton Waterproof pH Spear Pocket pH Tester
  • Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
  • WTW pH 1970i Heavy-Duty pH Meter
  • Oakton pH meter with probe
  • Oakton Waterproof pHTestr 30 Pocket pH Tester

Lab Scale Rotary Evaporator

Buy Lab Scale Rotary Evaporator Items Including:
  • 100-mL evaporator flask for Buchi Rotary Evaporators, 24/40 joint (YO-23123-62)
  • Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 115V (YO-23123-52)
  • Heating bath for Buchi Rotary Evaporators, holds up to 5L flask, 230VA (YO-23123-53).

Glass Door Lab Refrigerator

Buy Glass Door Lab Refrigerator Items Including:
  • Refrigerator, Blood Bank 52 cu ft with Casters, 2 Glass Doors, 115V/60 (NSBR522WWG/0)
  • Refrigerator, Blood Bank 52 cu ft with Casters, 2 Glass Doors, 230V/50 (NSBR522WWG/5).

Water Jet Aspirator Pumps

Buy Water Jet Aspirator Pumps Items Including:
  • Stainles Steel Aspirator for 35031-00 & -10, 1 Each (71121)
  • Cooling Coil for 35031-00 and -10 vacuum pumps (71501)
  • Filter Pack For 35031-00 & -10 Vacuum Pumps, 5 Pack (72200)
  • Filter Pack For 35031-00 & -10 Vacuum Pumps, 10 Pack (72210).

Stroboscope Experiment

Buy Stroboscope Experiment Items Including:
  • NIST-Traceable Calibration; Stroboscope (or strobe-type tachometers) (YO-17050-10)
  • Pocket-strobe™ Stroboscope
  • Portable LED Stroboscope, battery operated with 120 to 240VAC, 50/60 H
  • Stroboscope, Internal battery/115 VAC recharger, Kit (YO-87004-14).

Cellstar Co2 Incubator Replacement Parts

Buy Cellstar Co2 Incubator Replacement Parts Items Including:
  • Stacking kit for 5.4 cubic foot CO2 incubators (YO-39070-54).

Micro Flow Valve

Buy Micro Flow Valve Items Including:
  • Cole-Parmer Two-Way Micro Solenoid Rocker Valve, 1/4-28 UNF
  • Cole-Parmer Two-Way Micro Solenoid Rocker Valve, Manifold Mount
  • Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 1/4-28 UNF
  • Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 1/8 NPT(F).

Lab Grade Ph Meter

Buy Lab Grade Ph Meter Items Including:
  • Cole-Parmer Lab Dissolved Oxygen Meter, Digital
  • pH electrode with ATC
  • pH electrode, Gel Epoxy
  • Replacement Waterproof Combination pH Electrode
  • Gel-filled pH Electrode

Rainin Disp X Bottle Top Dispenser

Buy Rainin Disp X Bottle Top Dispenser Items Including:
  • Cole-Parmer Bottletop Dispenser .25-2.5 mL
  • Cole-Parmer Bottletop Dispenser .25-2.5 mL with 100 mL Glass Bottle
  • Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Glass Bottle
  • Cole-Parmer Bottletop Dispenser 5-60 mL with 1000 mL Glass Bottle.

Cole-Parmer Disposable Pipette Tips

Buy Cole-Parmer Disposable Pipette Tips Items Including:
  • Cole-Parmer Disposable Pipette Tips, sterile, 2" L, 0-200 µL, 960/c (A23091PRX)
  • Cole-Parmer Disposable Pipette Tips, sterile, 2" L, 0-200 µL, 960/c (A23071PRX)
  • Cole-Parmer Disposable Pipette Tips, non-sterile, 3-1/8" L, 200-1300.

Tungsten Carbide Microtome Blade

Buy Tungsten Carbide Microtome Blade Items Including:
  • TBS Tungsten-Carbide Edged Knife, 16 cm (6.3 in), “D” profile, with st
  • Tungsten-carbide blade (YO-04300-20).

Corning Vials

Buy Corning Vials Items Including:
  • Corning External Thread Cryogenic Vials; 2.0 mL, Self-Standing, 50/pac
  • Corning Internal Thread Cryogenic Vials; 2.0 mL, Washer Seal, Self-Sta
  • Corning Cryogenic Vial Cap Inserts; White, 500/case
  • Corning Cryogenic Vial Cap Inserts; Green, 500/case.

Laboratory Filtering Flasks

Buy Laboratory Filtering Flasks Items Including:
  • Thermo Scientific Nalgene polypropylene filtering flask, 1000 mL
  • Thermo Scientific Nalgene polypropylene filtering flask, 2000 mL
  • Labglass™ Filtering Flask; capacity, 1000-mL; 135mm OD x 225mm H
  • KIMAX filtering flask, 1000 mL.

Chemistry Lab Kits

Buy Chemistry Lab Kits Items Including:
  • Replacement reagents for basic soil chemistry test kit (R-5029)
  • Replacement reagents for deluxe soil chemistry test kit (R-5010-01)
  • General Lab Glassware Starter Kit.

Forma Scientific Co2 Incubator Repair Nashville

Buy Forma Scientific Co2 Incubator Repair Nashville Items Including:
  • Thermo Scientific Forma Water Jacketed CO2 Incubator; TC 23
  • Thermo Scientific Forma Water Jacketed CO2 Incubator; IR 12
  • Thermo Scientific Forma Water Jacketed CO2 Incubator; IR 23
  • Thermo Scientific Forma Water Jacketed CO2 Incubator; TC 12.

High Temperature Duct Tape

Buy High Temperature Duct Tape Items Including:
  • Heavily insulated fibrox heating tape, maximum exposure temperature: 9 (102080)
  • Heavily insulated samox heating tape, maximum exposure temperature: 14 (051100L)
  • Heavily insulated samox heating tape, maximum exposure temperature: 14 (052100).

2 Square Tube Fittings

Buy 2 Square Tube Fittings Items Including:
  • BARBED SANITARY CLAMP ADAPTER, 304L SS, 2" CLAMP X 2" ID TUBING (YO-30561-15)
  • Coupling Body, 1/2" OD Tube (HFC13812)
  • Coupling Body, Panel Mount, Valved, 1/2" OD Tube (HFCD12812)
  • Adapter,1/2" OD Tubing X NW 25 (C105-02-201).

Specific Gravity Pycnometer and Thermometer

Buy Specific Gravity Pycnometer and Thermometer Items Including:
  • H-B Instrument Specific Gravity Hydrometer Set, 8 Plain Form, 0.700 to.

Ph Meter Calibration Services

Buy Ph Meter Calibration Services Items Including:
  • NIST-Traceable Calibration; pH/mV Simulator (YO-17106-06)
  • Oakton pH 5+ meter with probe with NIST-traceable Calibration
  • Oakton pH 6+ meter with probe with NIST-traceable Calibration
  • Oakton advanced pH 110 meter only with NIST-traceable Calibration.

Model 05015 50 Laboratory Oven

Buy Model 05015 50 Laboratory Oven Items Including:
  • Dwyer Slacktube Manometer, Model 1211-50, 25 to 0 to 25 cm
  • Service Kit for Cole-Parmer Gear Pump Systems, Models 74013-50 and 55
  • Service kit for models 07135-50 through -57 (YO-07135-58)
  • DURAC , Bule Spirit, Verification Thermometer, 50/200C, Oven.

316 Stainless Steel Barbed Cross Connectors

Buy 316 Stainless Steel Barbed Cross Connectors Items Including:
  • Barbed Fittings; Cross Connector, 316 SS, 1/16" Tubing ID (YO-31208-51)
  • Barbed Fittings; Cross Connector, 316 SS, 3/32" Tubing ID (YO-31208-52)
  • Barbed Fittings; Cross Connector, 316 SS, 3/16" Tubing ID (YO-31208-54).

Molded Plastic Fume Hoods

Buy Molded Plastic Fume Hoods Items Including:
  • Gray fabric chair with black reinforced plastic base
  • Seat height 23" (5500-GRAY)
  • Royal blue fabric chair with black reinforced plastic base
  • Seat heig (5500-BLUE)
  • Sit-stand stool, 17"-22" height, black reinforced plastic base (3505).

Air Flow Meter Ppm

Buy Air Flow Meter Ppm Items Including:
  • Hydronic Manometer (HM680) Water/Air Flow Meter
  • Fluke 922 Air Flow / Digital Manometer (FLUKE-922/KIT)
  • Extech Portable Indoor Air Quality CO2 Meter
  • Extech Desktop Indoor Air Quality CO2 Monitor
  • Carbon Dioxide Meter with NIST-Traceable Certificate.



Hot Plate Stirrer Business & Industrial to Air Flow Meter Ppm

>