Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 21
Autoclave Sterilization Trays for Surgical Instruments
to
Burette Stand
Image
Product Detail
More Detail
Autoclave Sterilization Trays for Surgical Instruments
Buy Autoclave Sterilization Trays for Surgical Instruments Items Including:
Thermo Scientific Nalgene polypropylene pipette sterilizing tray with
Miltex ® Surgical Instrument Kit (YO-10918-81)
Cole-Parmer Autoclavable Draining Tray, PP (YO-06720-22)
Plain Gut Sterile Sutures, 19 mm, Needle: FS-2 3-0 27" (YO-10901-04).
More Details
Wall Mounted Digital Thermometer
Buy Wall Mounted Digital Thermometer Items Including:
Indoor/Outdoor Thermometer with Alarm (7141CP)
RTD Digital Meter, user Selectable C or F, plus wall mount and 4 meter (D1396-7)
Dual Channel Thermometer with 2 Wire Probes (4307CP)
Jumbo Refrigerator/freezer thermometer with Bottle Sensor (4227CP).
More Details
Nylon Mesh Filter Pore Size
Buy Nylon Mesh Filter Pore Size Items Including:
Cole-Parmer Cellulose Acetate Sterile Filters; pore size; 0.45µ; Ac
Cole-Parmer PTFE Sterile Syringe Filters; pore size; 0.45µ
Cole-Parmer PTFE Sterile Syringe Filters; pore size; 0.20µ
Cole-Parmer Nitrocellulose Filters; pore size; 0.20µ; 100/box.
More Details
Masterflex Norprene
Buy Masterflex Norprene Items Including:
Masterflex Norprene Food A60F Tubing, B/T 87, 25' (YO-06402-87)
Masterflex Norprene Food A60F Tubing, B/T 91, 25' (YO-06402-91)
Masterflex Norprene Food A60F Tubing, B/T 87, 10 FT (3.0 M)
Masterflex Norprene high-pressure tubing, L/S 15HP, 25 ft..
More Details
Silicone Rubber Beaker Heater Temperature Controller
Buy Silicone Rubber Beaker Heater Temperature Controller Items Including:
15-gallon flexible silicone rubber drum heater with thermostat: 50 to (DHCS21)
5-gallon flexible silicone rubber drum heater with thermostat: 50 to 4 (DHCS10)
5-gallon flexible silicone rubber drum heater with thermostat: 50 to 1 (DPCS20).
More Details
Disposable 45 Micron Syringe Filter
Buy Disposable 45 Micron Syringe Filter Items Including:
Advantec Disposable Syringe Filters, Mixed Cellulose Ester; 0.20 µm (YO-81054-24)
Advantec Disposable Syringe Filters, Mixed Cellulose Ester; 0.20 µm (YO-81054-26)
Advantec Disposable Syringe Filters, Mixed Cellulose Ester; 0.45 µm (YO-81054-28).
More Details
Luer Lock Port
Buy Luer Lock Port Items Including:
Polycarbonate individual manifolds with luer locks; 2 ports; 180 rotat (YO-06464-81)
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91)
Nickel Plated Brass Manifold, 4 male lock ports, 1 female luer port (YO-31507-92).
More Details
Pharmed Tubing
Buy Pharmed Tubing Items Including:
Tubing sets, PharMed BPT with 2 welded stops, 400 mm length, 0.25 mm I (SC0740)
2-Stop PharMed BPT Tubing, 0.25mm, 12/pack (95713-12)
2-Stop PharMed BPT Tubing, 0.51mm, 12/pack (95713-18)
2-Stop PharMed BPT Tubing, 0.89mm, 12/pack (95713-26).
More Details
Ebro Thermometers
Buy Ebro Thermometers Items Including:
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with genera (1340-5417)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with penetr (1340-5418)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with handle (1340-5416).
More Details
Love Temperature Controllers
Buy Love Temperature Controllers Items Including:
Love Temperature Control, 1/32 DIN, Universal Input, Voltage Pulse & R (32B-23)
Love Temperature Control, 1/32 DIN, Universal Input, Relay Outputs, Ra (32B-33)
Love Temperature Control, 1/16 DIN, Universal Input, Voltage Pulse & R (16B-23).
More Details
10 Ml Sample Vial
Buy 10 Ml Sample Vial Items Including:
2 mL sample vial Lab File, 288 cs
4 mL sample vial Lab File, 144cs
8 mL sample vial Lab File, 144cs
12 mL sample vial Lab File, 144 cs
16 mL sample vial Lab File, 144 cs
More Details
Dwyer Manometers
Buy Dwyer Manometers Items Including:
Dwyer Slacktube Manometer - 60" WC w/ 3/16" barb process connection
209:Incline Mnmtr.2/0/3 Inw Durablock Stat Inclined Manometer .2/0/3.0
Dwyer Series 490 Wet/Wet Handheld Digital Manometer 0 to 100 psi
Dwyer Type 475 Manometer, 100 psi.
More Details
Coors Ceramics
Buy Coors Ceramics Items Including:
Crucibles, Bitumen filter-, porcelain, autoclavable, Coors Ceramics Co (YO-17808-00)
Crucibles, unglazed porcelain, autoclavable, Coors Ceramics Company, 7 (YO-17809-00)
Dishes, evaporating, porcelain, glazed inside and partly outside, shal (60233).
More Details
Ptfe Stoppers
Buy Ptfe Stoppers Items Including:
Cole-Parmer® Cylindrical cuvettes with two stoppers of PTFE resin; (YO-83304-06)
Cole-Parmer® Cylindrical cuvettes with two stoppers of PTFE resin; (YO-83304-07)
Cole-Parmer® Cylindrical cuvettes with two stoppers of PTFE resin; (YO-83304-16).
More Details
Pipette Filters
Buy Pipette Filters Items Including:
epTIPS Filter Tips, 0.1 to 10 µL, medium (022491211)
Finntip Flex Filter Pipette Tips, 100-1000 uL, non-sterile, 1000/bag (94052440)
Ovation VistaRak ™ Micro Pipette Tips, 5 racks of 192 tips; Filter (4060-1332)
Filter non-sterile 5/pk (4580560).
More Details
Markson 88 Ph Meter
Buy Markson 88 Ph Meter Items Including:
Semi-dome, epoxy body, polygel, rugged bulb, waterproof, single-juncti (YO-35808-88)
Etatron DLXB pH/ORP Pump Control System, 2 L/hr, 10 bar max pressure, (DLXB-PH-RX/MBB) (YO-56022-88)
Cole-Parmer Extension Cable, 100 kΩ Thermistor, 50 ft.
More Details
3 Way Mechanical Vacuum Valves
Buy 3 Way Mechanical Vacuum Valves Items Including:
Three-way solenoid pinch valve; 12 VDC, 1/32" ID x 3/32" OD tubing
Three-way solenoid pinch valve; 24 VDC, 1/32" ID x 3/32" OD tubing
Three-way solenoid pinch valve; 12 VDC, 1/8" ID x 1/4" OD tubing (100P3MP-12-05).
More Details
Orion Electrode Storage Solution 810007
Buy Orion Electrode Storage Solution 810007 Items Including:
Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)
Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottl
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Oakton electrode storage solution.
More Details
Viton Valves
Buy Viton Valves Items Including:
Bellows metering pump with Viton valve and O-ring, 30 to 300 mL/min, 1
Nylon Miniature Check Valve, 1/8" hose barb, fluorosilicone diaphragm (YO-98553-00)
Nylon Miniature Check Valve, 1/4" hose barb, fluorosilicone diaphragm (YO-98553-02).
More Details
Ag Agcl Electrode Fill Solution
Buy Ag Agcl Electrode Fill Solution Items Including:
ROSS electrode fill solution, 3 M KCl
Case of five 60-mL bottles (810007)
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Filling solutions: use with Cl- electrodes (900017)
Filling solution f/ammonia ISE electrode (951202).
More Details
Threaded Pvc Pipes
Buy Threaded Pvc Pipes Items Including:
Pipe fittings, Bulkhead connectors, PVC, 1/4" NPT (F) x 27/32" OD1-3/8 (YO-06445-10)
Pipe fittings, Swivel bulkhead connector, PVC, 1" NPT (F),2-9/16", 4-3 (YO-31800-00)
PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV).
More Details
Cole-Parmer Thermocouple Thermometer
Buy Cole-Parmer Thermocouple Thermometer Items Including:
Cole-Parmer Thermocouple Thermometer with Data Output
Cole-Parmer Thermocouple Thermometer Type-K (4015CP)
Cole-Parmer Dual-Channel Thermometer with RS-232 Output
Cole-Parmer Dual-Channel Thermometer
Cole-Parmer Wide-Range Thermometer with Memory.
More Details
Ptfe Wash Bottles
Buy Ptfe Wash Bottles Items Including:
Thermo Scientific Nalgene Wash bottle, FEP, 125mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 250mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 500mL, 1/pk.
More Details
Ptfe Bottle
Buy Ptfe Bottle Items Including:
Cole-Parmer PTFE bottle, 100mL, 1/ea (015.100.2)
Cole-Parmer PTFE bottle, 150mL, 1/ea (015.150.2)
Cole-Parmer PTFE bottle, 250mL, 1/ea (015.250.2)
Cole-Parmer PTFE bottle, 500mL, 1/ea (015.500.2)
Cole-Parmer PTFE bottle, 1000mL, 1/ea (015.1000.2).
More Details
Controllers-Watlow Din-a-Mite Power Supply
Buy Controllers-Watlow Din-a-Mite Power Supply Items Including:
Din-a-mite power controller 18amp, 4.5 to 32Vdc input, 1 phase, 1 cont (DA10-24C0-0000)
Din-a-mite power controller 18amp, 4.5 to 32Vdc input, 1 phase, 1 cont (DA10-60C0-0000).
More Details
47 Mm Filter Paper
Buy 47 Mm Filter Paper Items Including:
Advantec PTFE Depth Filter, 2 µm pore, 47-mm diameter; pack of 10
Advantec PTFE Depth Filter, 4 µm pore, 47-mm diameter; pack of 10
Advantec PTFE Depth Filter, 5 µm pore, 47-mm diameter; pack of 20
Net filter, nylon, 11 µm, 47 mm dia (YO-29959-20).
More Details
Cole-Parmer Msds
Buy Cole-Parmer Msds Items Including:
Replacement MSDS Binder, 1-1/2", English/Spanish
Replacement MSDS Binder, 2-1/2", English
Zinc Formalin,1gal/ea
See MSDS for HAZ-MAT handling requirements unde
Lithium Blue,Buffered,8.0ph, 1gal/ea
More Details
Glass Cuvette
Buy Glass Cuvette Items Including:
Fluorimeter cuvettes; stopper top; glass; 3.5 mL capacity, 2/pack
Cole-Parmer Standard-Sized cuvettes; glass; 0.4 mL capacity
Cole-Parmer Standard-Sized cuvettes; glass; 10.5 mL capacity
Cole-Parmer Standard-Sized cuvettes; glass; 14.0 mL capacity.
More Details
British Pipe Thread Adapter
Buy British Pipe Thread Adapter Items Including:
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50)
Threaded adapter, stainless steel, 1/4" female NPT to 1/4" male BSPP (YO-31830-51).
More Details
Burette Stand
Buy Burette Stand Items Including:
Burette Clamp,single (18204)
Burette Clamp,double (18206)
Buret Support Stand, Porcelein Stand with Aluminum Support Rod
Buret Support Stand, Porcelein Stand with Stainless Steel Support Rod
Double Buret Clamp/Stand (916187).
More Details
70618
results
<<<Previous
17
18
19
20
21
22
23
24
25
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Autoclave Sterilization Trays for Surgical Instruments to Burette Stand
>
31125329-s-toc_2 31125329stoc2