Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 22
Glass Bottle Companies in Riverside California
to
Male Luer Lock Connector
Image
Product Detail
More Detail
Glass Bottle Companies in Riverside California
Buy Glass Bottle Companies in Riverside California Items Including:
Frio-temp Freezer Thermometer, -30/1c, 135mm Length, Bottle Filled Wi
Frio-temp Refrigerator Thermometer, -5/15c, 125mm Length, Bottle Fille (20700T)
Frio-temp Incubator Thermometer, 10/45c, 98mm Length, Bottle Filled Wi.
More Details
Nist Traceable Digital Thermometer
Buy Nist Traceable Digital Thermometer Items Including:
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with genera (1340-5417)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with penetr (1340-5418)
High-accuracy NIST-traceable Platinum RTD Mini-thermometer with handle (1340-5416).
More Details
5 Micron Nylon Filter
Buy 5 Micron Nylon Filter Items Including:
Filter Cartridge, Antimicrobial Pleated Polyester, 5 micron nominal-ra (HC/90-5-AMW)
Filter cartridge, "Big Blue", 20", carbon block, 5 micron (155583-43(EP-20BB))
Mini Air Filter, 5 micron filter, Max 49 SCFM, Max Pressure 150 PSIG,.
More Details
15 Gallon Carboy Hand Pump
Buy 15 Gallon Carboy Hand Pump Items Including:
Hand-operated threaded hand pump, 26 strokes/gallon, 3/4" NPT(F) adapt (MP1209)
Guzzler diaphragm hand pump, 8.5 GPM, 7 Strokes/gallon
Hand-operated siphon/lift pail pump, 32 strokes/gallon (9A)
Water Pump, 6 strokes/gallon, 3 foot inlet tube.
More Details
Pvdf Compression Fittings
Buy Pvdf Compression Fittings Items Including:
Compression fittings, Straight union, Kynar, 1/4" OD, 1", 5/8", 3/16"; (YO-30805-00)
Compression fittings, Straight union, Kynar,3/8" OD, 1-1/4", 13/16", 1 (YO-30805-02).
More Details
Cole-Parmer Pumps - Microfluidics
Buy Cole-Parmer Pumps - Microfluidics Items Including:
Replacement Tubing Set for Cole-Parmer 73160-XX pumps, Silicon, 0.8, 1
Replacement Tubing Set for Cole-Parmer 73160-XX pumps, Viton, 0.8, 1.6
Cole-Parmer Variable-Speed Peristaltic Tubing Pump - Ultra Low Flow of.
More Details
Chloride Selective Electrode
Buy Chloride Selective Electrode Items Including:
Cole-Parmer® Combination Ion Selective Electrodes, Chloride (Cl) (YO-27504-08)
ISE Standard solution, chloride, 0.1 M (YO-27503-08)
Thermo Scientific Orion ISE chloride electrode, solid-state, BNC conne (9417BN)
SOLUTION KIT CHLORIDE (YO-35802-62).
More Details
Tygon R 3603
Buy Tygon R 3603 Items Including:
TYGON LAB TUBING, Non-DEHP, 1/32"ID X 3/32"OD, 50 FT/PACK (YO-07407-70)
TYGON LAB TUBING, Non-DEHP, 1/2"ID X 11/16"OD, 50 FT/PACK (YO-07407-91)
TYGON LAB TUBING, Non-DEHP, 1/2"ID X 13/16"OD, 50 FT/PACK (YO-07407-93).
More Details
60 Ml Hdpe Bottle
Buy 60 Ml Hdpe Bottle Items Including:
Wide-Mouth Sample Containers, PP, 60 mL
60 mL Kimax Glass Dropping Bottle
Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, HDPE, 60 mL, 12 (YO-06035-11)
Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, HDPE, 60 mL, 72 (YO-06035-13).
More Details
Tygon B 44 4x
Buy Tygon B 44 4x Items Including:
Masterflex® Tygon® Food tubing (B 44-4X), L/S® 15, 50 ft
(YO-06419-15)
Masterflex® Tygon® Food tubing (B 44-4X), L/S® 24, 50 ft
(YO-06419-24)
Masterflex® Tygon® Food tubing (B 44-4X), L/S® 35, 50 ft
More Details
Temperature Indicator Strips
Buy Temperature Indicator Strips Items Including:
Temperature Labels, Refrigerator (YO-90308-80)
Temperature Labels, Freezer (YO-90308-85)
Temperature Labels, Cooler (YO-90308-90)
Fortyeight-point vertical thermometer with Magnetic Backing, 26 to 120 (YO-08078-48).
More Details
Autoclave Tray
Buy Autoclave Tray Items Including:
Thermo Scientific Nalgene polypropylene autoclaving tray, 1 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 2 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 3 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 5 L.
More Details
Nalgene Jerricans
Buy Nalgene Jerricans Items Including:
Thermo Scientific Nalgene high-density polyethylene jerrican with hand (2240-0015)
Thermo Scientific Nalgene high-density polyethylene jerrican with hand (2240-0025)
Thermo Scientific Nalgene high-density polyethylene jerrican with hand (2243-0013).
More Details
Microscope Cover Glass Circle 35mm
Buy Microscope Cover Glass Circle 35mm Items Including:
Cover glasses, Circular, 18-mm diameter, 0.13 to 0.17 mm thickness, ap
Cover glasses, 22 x 22 mm square, 0.16 to 0.19 mm thickness, approxima (22X22MM #1.5 48510-10)
Lab-Tek™ Chambered Cover Glass System; material, glass; number of w (YO-01838-10).
More Details
Antivibration Table
Buy Antivibration Table Items Including:
Marble Balance Table (TABLE).
More Details
Compressor Filters
Buy Compressor Filters Items Including:
Mini Air Regulator, Max 47 SCFM, 5 Micron Filter, Pressure range of 0
Combo Filter/Regulator and Lubricator, Max 44 SCFM, Max Pressure of 15
Inlet filter/silencer assembly, 1/4" NPT(M), 4 cfm, Paper
Compressed-air filter, 6 cfm (8A02N-OB2-BX).
More Details
1 Liter Laboratory Wash Bottles
Buy 1 Liter Laboratory Wash Bottles Items Including:
Thermo Scientific Nalgene PPCO wash bottle, 1 L
Thermo Scientific Nalgene Wash bottle, FEP, 125mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 250mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 1000mL, 1/pk
Chemware Wash Bottle PFA, 1 L.
More Details
Tds4tm 3000 Salt Salinity Tester
Buy Tds4tm 3000 Salt Salinity Tester Items Including:
Oakton SALT 6+ handheld salinity meter with probe
Oakton SALT 6+ handheld salinity meter with probe and NIST-traceable C
Oakton SALT 6+ handheld salinity meter kit with NIST-traceable Calibra
Oakton® Acorn™ SALT 6 Meter with probe (YO-35606-20).
More Details
Luer Fittings
Buy Luer Fittings Items Including:
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61).
More Details
Ptfe Compression Tube Fittings
Buy Ptfe Compression Tube Fittings Items Including:
Barbed fittings, Straight Connectors, 6mm, Chemflour® PTFE, each (YO-06361-10)
Compression reducing union, Pharmafluor™ PTFE, 1/4" OD x 1/8" OD (YO-06379-11)
Compress to male threaded adapter, Pharmafluor PTFE, 1/16" OD x 1/8" N.
More Details
50 Ml Pipettor
Buy 50 Ml Pipettor Items Including:
CORNING Stripette Serological Pipettes, 5.0 mL, Individually Wrapped P
Reagent Reservoir, 25 mL, 50/case, Individually wrapped, Sterile, Pyro
Reagent Reservoir, 25 mL, divided, 50/case, Individually wrapped, Ster.
More Details
250 Ml Plastic Burette
Buy 250 Ml Plastic Burette Items Including:
Cole-Parmer Bottletop Dispenser 1-10 mL with 250 mL Plastic Bottle
Pyrex Large Vol Burette 250 ml (2094-250)
Chemware Beaker PFA, 250 mL
Scienceware Tapered Polypropylene Beaker, 250 mL
Polypropylene graduated beaker with handle, 250 mL (V440941).
More Details
Thermo Scientific Ph Meters
Buy Thermo Scientific Ph Meters Items Including:
Thermo Scientific Orion PerpHecT pH buffer packs, 4.01, 10/PK
Thermo Scientific Orion PerpHecT pH buffer packs, 4.01, 25/PK
Thermo Scientific Orion PerpHecT pH buffer packs, 7.00, 10/PK
Thermo Scientific Orion PerpHecT pH buffer packs, 7.00, 25/PK.
More Details
50 Ml Test Tube
Buy 50 Ml Test Tube Items Including:
Pyrex Brand 9825 culture tube with screw cap; 50 mL, case of 192
Thermo Scientific Nunc Round-bottom test tubes; 50-mL; PPCO; 10/pack
Tubes, Volumetric, 100 mL, 50/pk
AIM600 Programmable Block Digestion System, 50 Place With 75 mL Tubes.
More Details
Mettler Toledo Ph Electrode Storage Solution
Buy Mettler Toledo Ph Electrode Storage Solution Items Including:
Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)
Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottl
Thermo Scientific Orion pH electrode cleaning solution A, 4 x 30 mL bo
Oakton electrode cleaning solution.
More Details
Sae Straight Thread Fittings Chart
Buy Sae Straight Thread Fittings Chart Items Including:
Straight threaded coupling, nylon, 1/8" NPT(F), pack of 6 (YO-06349-00)
Straight threaded coupling, nylon, 1/4" NPT(F), pack of 6 (YO-06349-01)
Straight threaded coupling, nylon, 3/8" NPT(F), pack of 6 (YO-06349-02).
More Details
2 Oz Amber Glass Bottles
Buy 2 Oz Amber Glass Bottles Items Including:
2000 mL Kimax Amber Media Bottles
30 mL Amber Glass Dropping Bottle, Glass dropper
30 mL Amber Glass Dropping Bottle, Plastic dropper
Cole-Parmer Bottle, Clear Boston Round, 2 oz, 24/case
Cole-Parmer Bottle, Graduated Medium Rounds, 2 oz, 48/case.
More Details
Stopcock Glass Burette 100ml
Buy Stopcock Glass Burette 100ml Items Including:
COLE PARMER BURETTE, 100ML WITH GLASS STOPCOCK
COLE PARMER BURETTE, 100ML WITH GLASS STOPCOCK, 4/CASE
Cole-Parmer Class B Burette w/PTFE stopcock; 10ML
Econ Burette Locking Stopcock (2116-LSO)
COLE PARMER BURETTE, 10ML WITH GLASS STOPCOCK, 6/CASE.
More Details
Female X 3 8 Barb
Buy Female X 3 8 Barb Items Including:
Quick-disconnect fittings, Straight-through hose barbs, PP, 3/8 (HFC17612)
Quick-disconnect fittings, Straight-through hose barbs, PP, 3/8 (HFC171012)
Quick-disconnect fittings, Straight-through hose barbs, PP, 3/8 (HFC171212).
More Details
Male Luer Lock Connector
Buy Male Luer Lock Connector Items Including:
Male luer lock plug, Nylon, 25/pk (YO-45505-56)
Male Luer Lock Plug, PP, 100 Per Pack (YO-45603-56)
Male luer with lock ring x female luer coupler, PP, 25/pk (YO-45500-80)
Male luer with lock ring x female luer coupler, Nylon, 25/pk (YO-45502-80).
More Details
70618
results
<<<Previous
18
19
20
21
22
23
24
25
26
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Glass Bottle Companies in Riverside California to Male Luer Lock Connector
>
31125329-s-toc_2 31125329stoc2