Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 23
Cole-Parmer Animal Feeding
to
Research Laboratory Supply Inc
Image
Product Detail
More Detail
Cole-Parmer Animal Feeding/Intubation Needles
Buy Cole-Parmer Animal Feeding/Intubation Needles Items Including:
Intubation tube-Straight 22g 1"straight 12/pk (YO-10919-20)
Intubation tube-Straight 22g 1-1/2"strt 12/pk (YO-10919-22)
Intubation tube-Straight 20g 1-1/2"strt 12/pk (YO-10919-24).
More Details
Pitot Tube Flow Meter
Buy Pitot Tube Flow Meter Items Including:
Fluke 922 Air Flow / Digital Manometer (FLUKE-922/KIT)
Brass Metering Valve with compression connections, 1/16" tube OD, 0.03 (1Z-NSL-BN-B-V)
Brass Metering Valve with compression connections, 1/8" tube OD, 0.031 (2Z-NSL-BN-B-V).
More Details
1000 Ml Hdpe Bottles
Buy 1000 Ml Hdpe Bottles Items Including:
Thermo Scientific Nalgene Centrifuge bottle; 1000-mL; PPCO; 4/pack (3120-1000)
Thermo Scientific Nalgene Centrifuge bottle; 1000-mL; PC; 4/pack (3122-1000)
Thermo Scientific Nalgene Centrifuge bottle; 1000-mL; PPCO; 4/pack (3120-1010).
More Details
Lamotte Turbidity Standards 2020
Buy Lamotte Turbidity Standards 2020 Items Including:
Turbidity standard, 10 NTU, for LaMotte 2020WE meter
Turbidity standard, 1000 NTU, for LaMotte 2020WE meter
Turbidity standard, 1000 NTU, for LaMotte 2020WI meter
LaMotte 2020we Handheld Turbidity Meter, EPA compliant.
More Details
High Temperature Circulators Fans
Buy High Temperature Circulators Fans Items Including:
High-Stability Immersion Digital Circulator; 120V; 50/60Hz (YO-01262-05).
More Details
Hach Sension Waterproof Ph/Mv Meter
Buy Hach Sension Waterproof Ph/Mv Meter Items Including:
Hach SensION™+ Wastewater pH Meter Kit
pH Electrode, Gel-filled for Portable meter
pH Electrode, Double-junction for Portable meter.
More Details
High Heat Resistant Gloves
Buy High Heat Resistant Gloves Items Including:
Ansell Thermaprene™ Heat-Resistant, Lined Neoprene Gloves, size 8,
Ansell Thermaprene™ Heat-Resistant, Lined Neoprene Gloves, size 10,
MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Medium
MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Extra-large.
More Details
Pyrex Beakers
Buy Pyrex Beakers Items Including:
Pyrex Brand 1000 Griffin Low-Form Beaker, 10 mL, 12/pk (1000-10 425120)
Pyrex Vista Griffin Style Beakers, 10mL, 12/pk (70000-10)
Pyrex Vista Griffin Style Beakers, 100mL 12/pk (70000-100)
Pyrex Vista Griffin Style Beakers, 150mL 12/pk (70000-150).
More Details
Oakton Ph Electrode Storage Solution Msds
Buy Oakton Ph Electrode Storage Solution Msds Items Including:
Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)
Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottl
Thermo Scientific Orion pH electrode cleaning solution A, 4 x 30 mL bo
Oakton electrode cleaning solution.
More Details
Parker Compression Fittings
Buy Parker Compression Fittings Items Including:
Compression fittings, male pipe adapter, straight, Black PP, 1/4" OD x (YO-06444-01)
Compression fittings, male pipe adapter, straight, Black PP, 3/8" OD x (YO-06444-02).
More Details
Hand Operated Vacuum Pump with Gauge
Buy Hand Operated Vacuum Pump with Gauge Items Including:
Hand-operated transfer pump for detergents, waxes, alkalis and certain
Hand-operated siphon pump, 5 GPM, Polyethylene (4005(LSS-1345))
Hand-operated siphon pump, 2 GPM, Polyethylene (4007)
Hand-operated drum pump, PVC body, 6 strokes/gallon, spout.
More Details
Orion Electrode Fill Solution Msds
Buy Orion Electrode Fill Solution Msds Items Including:
Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bot
Electrode fill solution, 4 M KCl with Ag+
Case of five 60-mL bottles (900011)
ROSS electrode fill solution, 3 M KCl
Case of five 60-mL bottles (810007).
More Details
Nalgene Spigot
Buy Nalgene Spigot Items Including:
Thermo Scientific Nalgene Replacement polypropylene cap, 24 mm
Thermo Scientific Nalgene Replacement polypropylene cap, 100 mm
Thermo Scientific Nalgene Replacement polypropylene cap, 53B
Thermo Scientific Nalgene Replacement, polypropylene spigot.
More Details
Hach Ph Buffer
Buy Hach Ph Buffer Items Including:
Turbidity Standards Kit for Hach 2100 N/AN Meters, 0, 20, 200, 1000, a (YO-08391-24)
Turbidity Standards Kit for Hach 2100P meters, 0, 20, 100, 800 NTU (YO-08391-60)
Turb Std, Hach 1720 Kit
Turb Standard 1.0, Hach 1720
Turb Standard 20, Hach 1720.
More Details
40 Micron Filter Paper Rolls
Buy 40 Micron Filter Paper Rolls Items Including:
Filter Cartridge, Pleated All-Polypropylene, 0.2 micron absolute-rated (HC-PP-40-0.2)
Filter Cartridge, Pleated All-Polypropylene, 0.45 micron absolute-rate (HC-PP-40-0.45)
Filter Cartridge, Pleated Polyester, 1 micron nominal-rated, 9-5/8" L.
More Details
1.5 Inch Sanitary Fitting
Buy 1.5 Inch Sanitary Fitting Items Including:
Polypropylene Sanitary Pipe Thread Adapter, 1 1/2" Clamp Size x 1 1/2" (YO-31806-15)
PVDF Sanitary Concentric Reducer, 2 1/2" x 1 1/2" Clamp Size (YO-31808-15)
ReSeal Sanitary Coupling, 3/4" ID, 1" OD, 3/4" Clamp.
More Details
Thin Wall Stainless Steel Beakers
Buy Thin Wall Stainless Steel Beakers Items Including:
Stainless-steel Griffin-style beaker with easy-pour rim, 125 mL (YO-07205-30)
Stainless-steel Griffin-style beaker with easy-pour rim, 250 mL (YO-07205-40)
Stainless steel Griffin-style beaker with easy-pour rim, 600 mL (YO-07205-50).
More Details
250 Ml Hdpe Bottles
Buy 250 Ml Hdpe Bottles Items Including:
Thermo Scientific Nalgene Centrifuge bottle; 250-mL; PC; 4/pack
Thermo Scientific Nalgene Centrifuge bottle; 250-mL; FEP; 1/pack
Cole-Parmer Bottletop Dispenser 1-10 mL with 250 mL Glass Bottle
Azlon solvent venting wash bottle for acetone, 250 mL.
More Details
Rainin Rt 20 Pipette Tips
Buy Rainin Rt 20 Pipette Tips Items Including:
0.1 to 20µL epTIPS pipette tips, Starter set (022491415)
20 to 300µL epTIPS pipette tips, Starter set (022491440)
Ovation VistaRak ™ Micro Pipette Tips, 5 racks of 192 tips; 0.2 to
Ovation VistaRak ™ Micro Pipette Tips, 5 racks of 192 tips, Steril.
More Details
Nalgene Fep Wash Bottle
Buy Nalgene Fep Wash Bottle Items Including:
Thermo Scientific Nalgene Wash bottle, FEP, 125mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 250mL, 1/pk
Thermo Scientific Nalgene Wash bottle, FEP, 500mL, 1/pk.
More Details
Pore Size on Filter Paper
Buy Pore Size on Filter Paper Items Including:
Glass Fiber Filters; 55mm dia; pore size, 1.6µ; thickness, 0.22mm; (GA5555MM)
Whatman Qualitative Filter Papers; 5.5 cm dia; pore size, 20-25µ; 1
Whatman Qualitative Filter Papers; 11.0 cm dia; pore size, 20-25 µ;.
More Details
Thin Wall Flexible Tubing
Buy Thin Wall Flexible Tubing Items Including:
Cole-Parmer PCR Tubes, individual, 0.2 mL, thin walled with dome caps,
PCR tubes, polypropylene, 0.2 ml, thin wall with flat caps, 1000/case
Tubing C-flex Braided; 3/4"id, 1 1/8"OD, 3/16" Wall, 15'.
More Details
Cylinder Clamp
Buy Cylinder Clamp Items Including:
Heavy Duty Bench Clamp
Bench Clamp with secure message (973036)
Bench Clamp with secure message (974036)
Replacement Bench Clamp
Stand (970040)
More Details
200 Ml Turbovap Tube Racks
Buy 200 Ml Turbovap Tube Racks Items Including:
Centrifuge Tube Rack for 50-mL centrifuge tubes (18794)
Centrifuge tube rack for 15-mL centrifuge tubes (YO-06696-02)
Test tube racks for 1.5 mL tube size; PP (YO-06703-40)
Floating Microcentrifuge Tube Rack (holds 20 X 0.5 mL Tubes) (F18875-1600).
More Details
Laboratory Brushes
Buy Laboratory Brushes Items Including:
Foam Laboratory Brush Set
Carboy Brush, 22", Nylon, 12/pk
Heavy Duty Carboy Brush, 22", Nylon, 12/pk
Heavy Duty Carboy Brush, 34", Nylon, 12/pk
Large Bottle/cylinder Brush, 20", 12/pk
More Details
John Guest Fittings Pack
Buy John Guest Fittings Pack Items Including:
Straight Unions, 1/4, 10 Per Pack (YO-34006-17)
Straight Unions, 3/8, 10 Per Pack (YO-34006-18)
Straight Unions, 1/2, 10 Per Pack (YO-34006-19)
Bulkhead Unions, 1/4, 10 Per Pack (YO-34006-20)
Bulkhead Unions, 3/8, 10 Per Pack (YO-34006-21).
More Details
Bspt Fittings
Buy Bspt Fittings Items Including:
Coupling Insert, Valved, 1/4" BSPT, 10/pack (APCD24004BSPT)
Coupling Insert, 1/4" BSPT, 10/pack (APC24004BSPT)
Barbed fitting kits, BSPT, adapters/elbows/tees, HDPE (YO-31514-32)
Coupling Insert, Non-Spill, 1/4" BSPT (NS4D24004BSPT).
More Details
Manifolds and Luer Lock
Buy Manifolds and Luer Lock Items Including:
Polycarbonate individual manifolds with luer locks; 2 ports; 180 rotat (YO-06464-81)
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91)
Nickel Plated Brass Manifold, 4 male lock ports, 1 female luer port (YO-31507-92).
More Details
4 Liter Amber Glass Bottles
Buy 4 Liter Amber Glass Bottles Items Including:
30 mL Amber Glass Dropping Bottle, Plastic dropper
60 mL Amber Glass Dropping Bottle, Plastic dropper
Bottle, precleaned, amber glass, narrow mouth, Qorpak, 60 mL, 20 mm ne
Bottle, precleaned, amber glass, narrow mouth, Qorpak, 120 mL, 22 mm n.
More Details
Research Laboratory Supply Inc
Buy Research Laboratory Supply Inc Items Including:
BIOSIMPLEX CARBOY EZ TOP ASSEMBLY, 20L, 2 PORTS, 3/8 X 5/8 TUBING, inc
BIOSIMPLEX CARBOY EZ TOP ASSEMBLY, 40L, 2 PORTS, 3/8 X 5/8 TUBING, inc
BIOSIMPLEX CARBOY EZ TOP ASSEMBLY, 20L, 3 PORTS, 3/8 X 5/8 TUBING, inc
Double Buret Clamp Stainless Steel.
More Details
70618
results
<<<Previous
19
20
21
22
23
24
25
26
27
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cole-Parmer Animal Feeding to Research Laboratory Supply Inc
>
31125329-s-toc_2 31125329stoc2