Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 2354

Cole Ra   to   Colde all-Plastic

Image
  Product Detail
More Detail

Cole Ra

Buy Cole Ra Items Including:
  • Rack insert, holds 150 culture tubes (to 105 mm); inserts on bottom ra (YO-98951-81)
  • Standard gear pump system with relief valve and 0.5 to 1.0 GPM flow ra
  • Filter Cartridge, Antimicrobial Pleated Polyester, 5 micron nominal-ra (HC/170-5-AMW).

Cole Hea

Buy Cole Hea Items Including:
  • Industrial Direct Insert Spring Loaded RTD Probe 4"L With Aluminum Hea (707A-100-F-4-3-316-AL-1/2-1-0).

Cole Ultra

Buy Cole Ultra Items Including:
  • Pump, High Precision - Ultra High Pressure Pumps, up to 18,000 psi, Co (U05SNX01)
  • Pump, High Precision - Ultra High Pressure Pumps, up to 18,000 psi, Co (F05SNXP1-SP1)
  • Ultra-Quiet Oil-lubricated Air Compressor, 4.5 cfm, 115 VAC (P150/50AL).

Cole Ext

Buy Cole Ext Items Including:
  • Belt Holder for Ext Probe Transducer TRA73
  • Wd-08516-30:Ext Cbl K 10FT Extention Cable Type K T/C (050697A-K).

Cole Microbiology

Buy Cole Microbiology Items Including:
  • Cole-Parmer Cetrimide Agar, 500 g
  • Cole-Parmer Azide Dextrose Broth, 500 g
  • Cole-Parmer Baird Parker Agar, 500 g
  • Cole-Parmer Blood Agar Base with Low pH, 500 g
  • Cole-Parmer Brain Heart Infusion Broth, 500 g

Cole Moistur

Buy Cole Moistur Items Including:
  • Cole-Parmer Extreme-Accuracy Thermometer 0.00°
  • Cole-Parmer Extreme-Accuracy Thermometer 0.00°, 25.00°, 37.00
  • Cole-Parmer Big-Digit 4-Alert Alarm Thermometer °C
  • Cole-Parmer Big-Digit 4-Alert Alarm Thermometer ° F
  • Cole-Parmer Monitoring Thermometer.

Fl1e H12rce

Buy Fl1e H12rce Items Including:
  • FL1E Base Unit (FL1E-H12RCE)
  • Starter Kit, Includes FL1E-H12RCE base unit, software, programming cab.

Dual Line

Buy Dual Line Items Including:
  • 1/16-DIN Temp
  • controller with dual-line display, SSRD, relay, 100-24 (940000000)
  • 1/16-DIN Temp
  • controller with dual-line display, relay, relay, 100-2 (941100000)
  • 1/16-DIN controller with dual-line display, RS-232, SSRD, relay, 100-2 (940 000 200).

Safet S

Buy Safet S Items Including:
  • Frio-temp Certified Verification Thermometer For Incubators, PFA Safet.

Mark 10

Buy Mark 10 Items Including:
  • Mark-10, EFCOMP-L, Complete options package for ESM301L Test Stand
  • Mark-10 AC1014 3/8" Jacobs Chuck
  • Mark-10 AC1006 Table Top Mounting Kit
  • Mark-10 G1016-2 Jacobs Chuck, 0.062", 0.375" Diameter
  • Mark-10 G1017 Bit Holder, 1/4" Hex.

Cole P-Xylene

Buy Cole P-Xylene Items Including:
  • XYLENES HISTO F-STYL POLY 1GAL (YO-88087-43)
  • ALPHA CHLORO P XYLENE 99+ 5ML (YO-88100-60)
  • 4-CHLORO-O-XYLENE 98% 5ML (YO-88167-44)
  • 4-BROMO-M-XYLENE 97% 10GR (YO-88270-56)
  • 5-IODO M XYLENE 99% 25ML (YO-88283-81)

Trans

Buy Trans
  • Polystyrene Trans-Boxes container; 6 sections, 1-5/8" x 6" x 1-1/2", 2
  • Polystyrene Trans-Boxes container; 18 sections, 1-5/8" x 1-7/8" x 1-1/
  • K-resin Trans-Boxes container; 24 sections, 2" x 2" x 2", 12/pack.

Quick Disconnect Small

Buy Quick Disconnect Small Items Including:
  • In-Line Coupling Body, pipe adapters, valved, 3/8" NPT(M), PP
  • (CQHD06100206)
  • Flared fitting, valved, 3/8" OD, PP, panel mount, each (CQHD06110106PANELMT)
  • Pipe adapter, valved, 3/8" NPT(M), PP, panel mount, each (CQHD06110206PANELMT).

Dispe Disp

Buy Dispe Disp Items Including:
  • Precision Turbine Flowmeter, 1.6 to 16 GPM, 1 1/2" sanitary, with disp (GSCP-075152-5)
  • Thermal Gas Mass Flow Controller, to 10 sccm Ar, Aluminum body w/ disp
  • Thermal Gas Mass Flow Controller, to 20 sccm Ar, Aluminum body w/ disp.

Cole Heavy

Buy Cole Heavy Items Including:
  • Heavy-duty Converter Clamp for 500- and 750-watt Ultrasonic Processors (YO-04710-94)
  • Heavy-Duty Support Assembly for 500- and 750-watt Ultrsonic Processor (830-00320)
  • Cole-Parmer Extension Cable, 10 ft
  • Cole-Parmer Extension Cable, 100Ω RTD, 10 ft.

Safe T Guard

Buy Safe T Guard Items Including:
  • Taylor 9842FDA Safe-T-Guard Pocket Thermometer with Antimcrobial Sleev
  • Taylor 9848EFDA Pen-Style Safe-T-Guard Pocket Thermometer with Antimcr
  • Taylor 9847N Pen-Style Waterproof Digital Thermometer with Anti-Microb.

Fixed Ga

Buy Fixed Ga Items Including:
  • Sperian Biosystems NXP Fixed-Mount Gas Monitor, Replacement Oxygen Sen
  • Bacharach GDC-350 Fixed-Mount Gas Monitor, Carbon monoxide
  • Bacharach GDC-350 Fixed-Mount Gas Monitor, Nitrogen dioxide
  • Bacharach GDC-350 Fixed-Mount Gas Monitor, Sulfur dioxide.

Gear Driven

Buy Gear Driven Items Including:
  • Ismatec™ low-speed planetary gear-driven pump, 4 channels, 115/230
  • Ismatec™ standard-speed planetary gear-driven pump, 4 channels, 115.

Photo Sensitive

Buy Photo Sensitive Items Including:
  • Whirl-Pak Photo-Sensitive Bag 18 oz (YO-06498-06)
  • Amber Bag-Seal Top 2"W x 3"L
  • Amber Bag-Seal Top 3"W x 5"L
  • Amber Bag-Seal Top 4"W x 6"L
  • Amber Bag-Seal Top 5"W x 8"L

End-to-End C

Buy End-to-End C Items Including:
  • GE ROSave.Z Depth Cartridge Filter, 5.0 micron, plain end, 10" (1236581)
  • GE ROSave.Z Depth Cartridge Filter, 5.0 micron, plain end, 20" length (1236263)
  • GE ROSave.Z Depth Cartridge Filter, 1.0 micron, plain end, 20" length (1236262).

Cole Al

Buy Cole Al Items Including:
  • Industrial Direct Insert RTD Probe With Aluminum Head, 6" Length (707A-100-F-6-3-316-AL)
  • Palcam Listeria agar according to van Netten et al., 500g (YO-36152-54)
  • Safety Sign, Danger Lockout Tagout Equipment Before Entering, 10x14 Al (MLKT105VA).

Cole Controllers-Benchtop

Buy Cole Controllers-Benchtop Items Including:
  • Standard four-circuit programmable timer/controller, benchtop with dry (XT-4S-240V)
  • Deluxe four-circuit programmable timer/controller, benchtop with outle (XT-4N8)
  • Deluxe four-circuit programmable timer/controller, benchtop with outle (XT-4N8-240V).

Grade Surgical-Grade

Buy Grade Surgical-Grade Items Including:
  • Operating scissors, Dissecting grade, Sharp point (YO-10918-06)
  • Operating scissors, Dissecting Grade, Blunt point (YO-10918-08)
  • Halsted mosquito forceps, Dissecting-grade, Curved (YO-10918-16)
  • Adson Dressing Forceps, Dissecting-grade (YO-10918-22).

Cole 12&

Buy Cole 12& Items Including:
  • Cole-Parmer Battery-powered Digital Gauge, 0 to 15.00 PSI; 1/4" NPT(M)
  • Cole-Parmer 6-watt UV Lamp with 302nm Wavelength and White Light Tubes (YO-97620-12)
  • Very-Low Differential Transducer; 0 to 25" Water ~ 4 to 20 mA Output (YO-98073-12).

Cole Flo

Buy Cole Flo Items Including:
  • Flowmeter, 30-300 mL/min (YO-32550-01)
  • Flowmeter 100 - 1000 mL/min (YO-32550-03)
  • Flowmeter 200 - 2000 mL/min (YO-32550-05)
  • Flowmeter 300 - 3000 mL/min (YO-32550-07)
  • Flowmeter 500 - 5000 mL/min (YO-32550-09)

B & K Bk P

Buy B & K Bk P Items Including:
  • Disposable polypropylene Griffin low-form beakers, 100mL 100/cs (BMP-BK-100)
  • Disposable polypropylene Griffin low-form beakers, 100mL 1000/cs (BMP-BK-100)
  • Disposable polypropylene Griffin low-form beakers, 150mL 100/cs (BMP-BK-150).

1 M Tris

Buy 1 M Tris Items Including:
  • Silicone Gasket, 100 MESH, Size 1-1/2" Tri-Clamp (YO-30523-86)
  • Viton Gasket, 10 MESH, Size 1-1/2" Tri-Clamp (YO-30536-06)
  • Viton Gasket, 20 MESH, Size 1-1/2" Tri-Clamp (YO-30536-26)
  • Viton Gasket, 100 MESH, Size 1-1/2" Tri-Clamp.

Colde all-Plastic

Buy Colde all-Plastic Items Including:
  • Rotary Evaporator, Safety-Coated Dry Ice Condenser, Advanced Controlle (YO-23012-26)
  • Rotary Evaporator, Safety-coated Dry Ice Condenser, Professional Contr (YO-23012-27)
  • Rotary Evaporator, Safety-Coated Dry Ice Reflux Condenser, 220V (YO-23012-45).

70618 results


Cole Ra to Colde all-Plastic

>