Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 24

Inch Barb Check Valve   to   Stainless Steel Tubing Split Ring Hanger

Image
  Product Detail
More Detail

Inch Barb Check Valve

Buy Inch Barb Check Valve Items Including:
  • PP Check Valve with built-in screen, 3/8" hose barb (YO-07091-55)
  • Nylon Miniature Check Valve, 1/8" hose barb, fluorosilicone diaphragm (YO-98553-00)
  • Nylon Miniature Check Valve, 1/4" hose barb, fluorosilicone diaphragm (YO-98553-02).

4 Inch Plastic Bulkhead Fittings

Buy 4 Inch Plastic Bulkhead Fittings Items Including:
  • BARBED ELBOW, 3/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-75)
  • BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
  • Barbed Fittings; Cross Connector, Clear PP, 1/4-inch Tubing ID, 50/pk (YO-31417-11).

Filter Paper 0.45 Microns

Buy Filter Paper 0.45 Microns Items Including:
  • Filter Cartridge, Pleated All-Polypropylene, 0.45 micron absolute-rate (HC-PP-40-0.45)
  • Filter Cartridge, Pleated All-Polypropylene, 0.45 micron absolute-rate (HC-PP-170-0.45)
  • Gridded Membrane Filters, Sterile, Green/dark Green, 0.45 Micron.

Nalgene Spigot Caps

Buy Nalgene Spigot Caps Items Including:
  • Thermo Scientific Nalgene filling/venting cap for 1/2" tubing, 83B
  • Thermo Scientific Nalgene filling/venting cap for 1/4" tubing, 53B
  • Thermo Scientific Nalgene filling/venting cap for 1/4" tubing, 83B.

Cole-Parmer Fep Tubing

Buy Cole-Parmer Fep Tubing Items Including:
  • FEP Tubing, 3/8"ID x 7/16"OD, 25 ft/pack (YO-06406-12)
  • FEP Tubing, 1/2"ID x 9/16"OD, 25 ft/pack (YO-06406-18)
  • FEP Tubing, 1/32"ID x 1/16"OD, 25 ft/pack (YO-06406-60)
  • FEP Tubing, 5/8"ID x 11/16"OD, 25 ft/pack.

Siliconized Glass Vials

Buy Siliconized Glass Vials Items Including:
  • Glass Vials PTFE/SILiCone SEPTA Clear 12 ml 200/pk (B7990-12)
  • Glass Vials PTFE/SILiCone SEPTA Clear 16 ml 200/pk (B7990-4)
  • Glass Vials PTFE/SILiCone SEPTA Clear 22 ml 200/pk (B7990-5)
  • Glass Vials PTFE/SILiCone SEPTA Amber 12 ml 200/pk (B7990-12A).

50 Micron Filter

Buy 50 Micron Filter Items Including:
  • Filter Cartridge, Antimicrobial Pleated Polyester, 50 micron nominal-r (HC/40-50-AM)
  • Filter Cartridge, Antimicrobial Pleated Polyester, 50 micron nominal-r (HC/170-50-AM)
  • GE ROSave.Z Depth Cartridge Filter, 5.0 micron, plain end, 10" (1236581).

Npt to 1 4 Barb Fitting

Buy Npt to 1 4 Barb Fitting Items Including:
  • Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 1/4" I (YO-30610-38)
  • Barbed fittings, NPT male branch tees, Natural PP, 1/8" NPT x 1/4" ID, (YO-30610-72).

Tank Spray Guns

Buy Tank Spray Guns Items Including:
  • Spray Gun PTFE Di H20 3/8" (THG-DN6P-TFEV)
  • Spray Gun PTFE Di H20 1/2" (THG-DN8P-TFEV)
  • Spray Gun Recirculating 3/8" (THG-DR6P-TFE)
  • Spray Gun Recirculating Assmby (THG-DR6PVHE02)
  • Dispensing Gun PTFE 3/8"npt (THG-LD6P-TFEV).

Nist Traceable Non Mercury Immersion Thermometer

Buy Nist Traceable Non Mercury Immersion Thermometer Items Including:
  • Easy-read NIST Certified Thermometer, -20/110°C, 300mm Length, 76m (30506)
  • Easy-read NIST Certified Thermometer, -20/110°C, 300mm Length, Tot (30500C)
  • Easy-read NIST Certified Thermometer, -10/225c, 355mm Length, Total Im (30520C).

Cole-Parmer Stainless Steel Push-to-Connect Fittings

Buy Cole-Parmer Stainless Steel Push-to-Connect Fittings Items Including:
  • Push-to-connect male threaded adapter, 316 stainless steel, 1/4" x 1/8 (YO-31370-00)
  • Push-to-connect union elbow, 316 stainless steel, 1/2" (YO-31370-21)
  • Push-to-connect union tee, 316 stainless steel, 1/4" (YO-31370-22).

Oakton Conductivity Standard Solution

Buy Oakton Conductivity Standard Solution Items Including:
  • Oakton conductivity solution, 12,880 µS
  • Oakton® conductivity solution, 1413 µS
  • Oakton® conductivity solution, 2764 µS
  • Oakton® conductivity solution, 2070 µS
  • Oakton® conductivity solution, 80,000 µS

Kynar Fittings for Tubing

Buy Kynar Fittings for Tubing Items Including:
  • Barbed fitting kits, Threaded elbow/tee kits, Kynar, fittings kits
  • Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
  • Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62).

Nalgene Replacement Cap

Buy Nalgene Replacement Cap Items Including:
  • Thermo Scientific Nalgene Replacement polypropylene cap, 24 mm
  • Thermo Scientific Nalgene Replacement polypropylene cap, 28 mm
  • Thermo Scientific Nalgene Replacement polypropylene cap, 53B
  • Thermo Scientific Nalgene Replacement polypropylene cap, 83B.

Full Lens Magnifying Safety Glasses

Buy Full Lens Magnifying Safety Glasses Items Including:
  • U.S.SAFETY® SOLARIS Safety Glasses, Charcoal gray frame, Clear len (97101)
  • 3M Virtua™ 1.5 Magnifier Safety Eyewear, Clear Lens
  • 3M Virtua™ 2.0 Magnifier Safety Eyewear, Clear Lens
  • Uvex Protege™ Safety Glasses, Black frame, Clear lens, Ultra-dura c.

Etfe Tubing

Buy Etfe Tubing Items Including:
  • Chemfluor ETFE tubing, 1/50 x 1/16", 50 ft/pk (YO-06488-07)
  • Chemfluor ETFE tubing, 0.016 x 1/16", 50 ft/pk (YO-06488-08)
  • Chemfluor ETFE tubing, 0.232 x 5/16", 50 ft/pk (YO-06488-13).

Pipettor Stands

Buy Pipettor Stands Items Including:
  • Oxford Plexiglas pipettor rack
  • Double-bin pipettor workstation
  • Acrylic pipettor caddy
  • Pipettor stand (705385)
  • Eppendorf Research™ pro Electronic Pipettor, Charging Stand, for On

Connection 1/4" Npt(F)

Buy Connection 1/4" Npt(F) Items Including:
  • Plug Valve with Pipe Thread Connections, 1/4" NPT(F) (D1071794)
  • PVC Check Valve, 1/4" NPT(F) connections (CKD025V-PV)
  • PP Check Valve, 1/4" NPT(F) connections (CKD025V-PP)
  • PTFE Check Valve, 1/4" NPT(F) connections (CKD025V-TF).

Ceramic Furnaces

Buy Ceramic Furnaces Items Including:
  • Ceramic Ribbed Plate For Furnace Model Lt 3 (691600507)
  • Ceramic Ribbed Plate For Furnace Model Lt 24 (691600874)
  • Ceramic hearth plate for 130 cu-in furnaces (9353053)
  • Ceramic Hearth Plate for 1750 cu in Furnace with Single Set Point Cont (9353060).

Light Intensity Meters

Buy Light Intensity Meters Items Including:
  • UVX Digital UV Intensity Meter (97-0015-02(UVX))
  • UV Intensity Meter Sensor, 365 nm (97-0016-02(UVX36))
  • UV Intensity Meter Sensor, 302 nm (97-0016-04(UVX31))
  • Extech High-Intensity, Dual-Scale Light Meter
  • Attenuator, 10:1 (98-0035-01).

Natural Polypropylene Fittings

Buy Natural Polypropylene Fittings Items Including:
  • Barbed fitting kits, Threaded elbow/tee kits, Natural PP, fittings kit (YO-41514-27)
  • Y-connectors, Polypropylene, 3/4", Pack Of 5 (Y0-12NP (5 PACK))
  • Barbed fittings, Straight Connector, Natural PP, 1/4" ID, 1/8", 1-31/3 (YO-30610-00).

Wide Mouth Hdpe Bottles

Buy Wide Mouth Hdpe Bottles Items Including:
  • Thermo Scientific Nalgene Wide-Mouth Economy Bottle, HDPE, 30 mL, 12/p (YO-06035-31)
  • Thermo Scientific Nalgene Wide-Mouth Economy Bottle, HDPE, 30 mL, 72/p (YO-06035-33).

Lab Disc Magnetic Stirrer

Buy Lab Disc Magnetic Stirrer Items Including:
  • Ika Ultra-Flat Lab Disc Stirrer 230V (3277700)
  • Ika Ultra-Flat Lab Disc Stirrer Sunny Side Up (3362300)
  • Ika Ultra-Flat Lab Disc Stirrer Orange (3578000)
  • Ika Ultra-Flat Lab Disc Stirrer Green (3578200)
  • Ika Ultra-Flat Lab Disc Stirrer Pink (3578400).

Large Volume Pipette

Buy Large Volume Pipette Items Including:
  • Mla™ D-tipper Large Tip Pipette, Red, 200µL
  • 10 mL Cole-Parmer large-tip opening serological pipette (YO-25562-26)
  • 25 mL Cole-Parmer large-tip opening serological pipette (YO-25562-28)
  • Mla™ Precision Pipette, Large Tip, Red, 200µL.

Nalgene Products

Buy Nalgene Products Items Including:
  • Thermo Scientific Nalgene Replacement polypropylene cap, 24 mm
  • Thermo Scientific Nalgene Replacement polypropylene cap, 28 mm
  • Thermo Scientific Nalgene Replacement polypropylene cap, 53B
  • Thermo Scientific Nalgene Replacement polypropylene cap, 83B.

Dräger Pac 3500

Buy Dräger Pac 3500 Items Including:
  • Drager PAC 3500 Single-Gas Monitors, O2
  • Drager PAC 3500 Single-Gas Monitors, CO
  • Drager PAC 5500 Single-Gas Monitors, O2
  • Drager PAC 5500 Single-Gas Monitors, CO
  • Replacement Hydrogen Sulfide Sensor, 0 to 100 ppm

Plastic Graduated Beakers with Handles

Buy Plastic Graduated Beakers with Handles Items Including:
  • Polypropylene graduated beaker with handle, 1000 mL (V442941)
  • Polypropylene graduated beaker with handle, 2000 mL (V443941)
  • Polypropylene graduated beaker with handle, 3000 mL (V444941)
  • Polypropylene graduated beaker with handle, 5000 mL (V445941).

Hamilton Microliter Low-Volume Syringes with Needle

Buy Hamilton Microliter Low-Volume Syringes with Needle Items Including:
  • Hamilton Microliter Microsyringes, 0.5 µL, 25 gauge
  • Hamilton Microliter Microsyringes, 1.0 µL, 25 gauge
  • Hamilton Microliter Microsyringes, 2.0 µL, 25 gauge
  • Hamilton Microliter Microsyringes, 5.0 µL, 24 gauge.

Water Quality Iron

Buy Water Quality Iron Items Including:
  • CHEMets Colorimetric Iron Test Kit
  • Aqua Comparator Test Kit, Iron, 1-10 mg/L (L147320)
  • Iron TPTZ Reagent Powder Pillow Packs, 10 mL, 100 Tests (RP055-0)
  • Hach Total Iron (USEPA) Test Kit (21057-69)
  • Orbeco-Hellige SC400 Colorimeter, Iron (SC400-70).

Stainless Steel Tubing Split Ring Hanger

Buy Stainless Steel Tubing Split Ring Hanger Items Including:
  • 316 SS Vacuum tubing, NW 16 flange, 10"
  • 316 SS Vacuum tubing, NW 16 flange, 20"
  • 316 SS Vacuum tubing, NW 16 flange, 40"
  • Clamp, ISO-K, Stainless Steel, each (C100-07-156)
  • Adapter, NW 16 to 1/4" Tube OD , 316L SS (C105-02-101).



Inch Barb Check Valve to Stainless Steel Tubing Split Ring Hanger

>