Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 25
Q Tips Swabs
to
Specific Gravity Bottles Pycnometer 50ml
Image
Product Detail
More Detail
Q Tips Swabs
Buy Q Tips Swabs Items Including:
Cleanroom swab; fabric with knit polyester tip and sealed polypropylen (TX759B)
Cleanroom swab; foam bud tip and hardwood handle; 1.10"L tip, 500/pk (TX700B)
Cleanroom swab; circular-foam tip and polypropylene handle; 0.81"L tip (TX708A).
More Details
Fluke Thermocouple Thermometer 51 52 Ii
Buy Fluke Thermocouple Thermometer 51 52 Ii Items Including:
Fluke 50-Series II (Model 51) Thermocouple Thermometer with Single In (FLUKE-51-2 60HZ)
Fluke 50-Series II (Model 52) Thermocouple Thermometer with Dual Inpu (FLUKE-52-2 60HZ).
More Details
8 Oz Amber Glass Jars
Buy 8 Oz Amber Glass Jars Items Including:
Thermo Scientific Nalgene multipurpose amber polyetherimide jar, 2 1/8
Kimble Straight-Sided Jar, Amber, 8 oz, 12/cs
Bottle, precleaned, amber glass, narrow mouth, Qorpak, 60 mL, 20 mm ne
Cole-Parmer Bottle, Amber Boston Round, 8 oz, 24/case.
More Details
Sterile Luer Lock Connector
Buy Sterile Luer Lock Connector Items Including:
Male luer lock plug, Nylon, 25/pk (YO-45505-56)
B-D™ Disposable Syringes, Luer-Lock Tips, 10 mL (YO-07940-12)
B-D™ Disposable Syringes, Luer-Lock Tips, 30 mL (YO-07940-20)
60 mL B-D plastic disposable syringe, luer-lock tip, 40 / box (YO-07940-30).
More Details
Extra Long Nitrile Gloves
Buy Extra Long Nitrile Gloves Items Including:
Ansell Touch-N-Tuff™ Nitrile Gloves, Powder-free, Extra-large
Microflex Nitron One Nitrile Gloves, extra-large
Kimberly-Clark Nitrile Gloves, 9.5" length, Extra-large; 90/box
Kimberly-Clark Nitrile gloves, 12" length, Extra-large; 50/box.
More Details
Coliform Test Kit for Water Testing
Buy Coliform Test Kit for Water Testing Items Including:
Coliform Test Kit (4-3616)
Readycult Coliforms snap pack, 20/pk (YO-36152-10)
Readycult Coliforms snap pack, 200/pk (YO-36152-12)
Sterile Sample Bottle with Dechilorinating agent, 345/cs (YO-36152-16).
More Details
Thermo Bag
Buy Thermo Bag Items Including:
Pipette Tips, Finntip-Flex, 5-300uL, 1000/bag (94060520)
Pipette Tips, Finntip-Flex, 1-10mL, 100/BAG (94060970)
Finntip Flex Filter Pipette Tips, 1-10 mL, 50/bag (94056970)
Finntip Flex Filter Pipette Tips, 5-100uL, non-sterile, 100/bag (94052210).
More Details
Mini Pumps
Buy Mini Pumps Items Including:
Masterflex platinum-cured silicone sanitary tubing with 1/2" mini-conn (YO-96100-26)
Mini Air Cooled Magnetic Drive Centrifugal Pump, 2.3 GPM or 4.6 FT, 3/ (YO-07105-01)
Mini Lubricator with bowl guard, Max 32 SCFM, Max Pressure of 150 PSI,.
More Details
To 5 16 Barb Reducer
Buy To 5 16 Barb Reducer Items Including:
Barbed fittings, Reducing Connector, Natural PP, 5/8" x 5/16" ID, 3/16 (YO-30610-14)
Reducing Connector, Polyethylene, 5/8" x 5/16", 10 PACK (YO-30621-98)
Reducing Connector, Nylon, 5/8" x 5/16", 10 PACK (YO-30621-99).
More Details
250 Ml Centrifuge Tube Rack
Buy 250 Ml Centrifuge Tube Rack Items Including:
Centrifuge Tube Rack for 50-mL centrifuge tubes (18794)
Centrifuge tube rack for 15-mL centrifuge tubes (YO-06696-02)
Thermo Scientific Nalgene Centrifuge bottle; 250-mL; PPCO; 4/pack
Thermo Scientific Nalgene Centrifuge bottle; 250-mL; PC; 4/pack.
More Details
Sanitary Fittings Npt Adapter
Buy Sanitary Fittings Npt Adapter Items Including:
Polypropylene Sanitary Pipe Thread Adapter, 1 1/2" Clamp Size x 1" NPT (YO-31805-69)
Polypropylene Sanitary Pipe Thread Adapter, 2" Clamp Size x 1 1/4" NPT (YO-31805-78).
More Details
Asco Solenoid Valve Distributor
Buy Asco Solenoid Valve Distributor Items Including:
ASCO Solenoid Valve, Two-Way Normally Closed, 12V, 0.65 Watts, 100 psi
ASCO Solenoid Valve, Two-Way Normally Closed, 24V, 0.65 Watts, 100 psi
ASCO Solenoid Valve, Three-Way Universal Operation, 12V, 0.65 Watts, 1.
More Details
Ptfe Compression Fittings
Buy Ptfe Compression Fittings Items Including:
Compression reducing union, Pharmafluor™ PTFE, 1/4" OD x 1/8" OD (YO-06379-11)
Straight compression union, Pharmafluor™ PTFE, 3mm tubing OD (YO-06396-12)
Compression Etfe Nut And PTFE Ferrule Assembly For pHarmafluor Fitting (YO-31001-69).
More Details
Troemner Gas Cylinder Securing Safety Straps
Buy Troemner Gas Cylinder Securing Safety Straps Items Including:
Bench Clamp with secure message (973036)
Wall Bracket with secure message (973037)
Bench Clamp with secure message (974036)
Wall Bracket with secure message (974037)
Safety Strap without Message
More Details
Neoprene Stoppers
Buy Neoprene Stoppers Items Including:
Green neoprene stopper, size 000
Green neoprene stopper, size 00
Green neoprene stopper, size 0
Green neoprene stopper, size 1
Green neoprene stopper, size 2
More Details
Female Luer Fitting Solid Works
Buy Female Luer Fitting Solid Works Items Including:
Cole-Parmer Animal Free Female Luer Fittings, 3/16", PP, 25/pk
Cole-Parmer Animal Free Female Luer Fittings, PP, 25/pk
Adapters with caps; female luer fitting; Tefzel
3-way "Y" adapter, 1 female luer, 2 male luer lock (YO-31507-81).
More Details
High Temperature Plastic Tube Fittings
Buy High Temperature Plastic Tube Fittings Items Including:
Zetex High-Temperature Gloves, up to 1100°F, one size fits all, 14
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62).
More Details
Type K Thermocouple Probes
Buy Type K Thermocouple Probes Items Including:
Type J All stainless steel thermocouple probe; 8"L (050797A-J)
Type K All stainless steel thermocouple probe; 8"L (050797A-K)
Type J High Temperature 12"L, .040" diameter thermocouple probe with s (073098D-J-G-12).
More Details
Conductivity Standards
Buy Conductivity Standards Items Including:
Cole-Parmer conductivity standard, 1000 µS, 16 ounces
Cole-Parmer conductivity standard, 100,000 µS, 16 ounces
Cole-Parmer conductivity standard, 1 µS, 16 ounces, individually te
Cole-Parmer conductivity standard, 5 µS, 16 ounces, individually te.
More Details
125 Ml Hdpe Bottles
Buy 125 Ml Hdpe Bottles Items Including:
Thermo Scientific Nalgene Centrifuge bottle; 250-mL; HDPE; 6/pack
Thermo Scientific Nalgene wide-mouth Unitary wash bottle, 125 mL
Scienceware wash bottle, 125 mL
Thermo Scientific Nalgene Narrow-Mouth Economy Bottle, HDPE, 125 mL, 1 (YO-06035-15).
More Details
Small Barb Fittings
Buy Small Barb Fittings Items Including:
Barbed fitting kits, Barbed connector/reducer kits, PP, small fittings (YO-31514-03)
Barbed fitting kits, Threaded barb adapter kits, Natural PP, small fit (YO-31514-11)
Barbed fitting kits, Barbed connector/reducer kits, Kynar, small fitti.
More Details
Luer Lock
Buy Luer Lock Items Including:
Syringes with PTFE luer lock; syringe capacity; 50µL; needle gauge; (YO-07939-32)
Syringes with PTFE luer lock; syringe capacity; 100µL; needle gauge (YO-07939-33)
Syringes with PTFE luer lock; syringe capacity; 1 mL; needle gauge; 22 (YO-07939-80).
More Details
Cole-Parmer Low-Pressure-Drop Liquid Flow Switches
Buy Cole-Parmer Low-Pressure-Drop Liquid Flow Switches Items Including:
Normally open flow switch for liquids; 0.5 GPM (YO-32776-00)
Normally open flow switch for liquids; 1.0 GPM (YO-32776-01)
Normally open flow switch for liquids; 2.0 GPM (YO-32776-02)
Normally open flow switch for liquids; 3.0 GPM (YO-32776-03).
More Details
1 Mm Id Ptfe Tubing
Buy 1 Mm Id Ptfe Tubing Items Including:
Assembly Of Flared 0.040 X 0.066" PTFE Tubing, 36", With 1/4-28 Fittin (YO-96154-03)
Assembly Of Flared 0.062 X 0.125" PTFE Tubing, 24", With 1/4-28 Fittin (YO-96154-15)
Denatured PTFE tubing, 0.086 x 1/8, 25' per pack (TID01-8).
More Details
Unf Pipe Fittings
Buy Unf Pipe Fittings Items Including:
Pipe fittings, Nipples UNF(M), Kynar, 10-32 UNF, 13/32", 5/16", 3/32"; (YO-30804-95)
Pipe fittings, Nipple, Nylon, 10-32 UNF, 3/8", 1/4", 1/16"; 25/pack (YO-31502-10).
More Details
Microliter Syringes
Buy Microliter Syringes Items Including:
Standard Microliter Syringes, 10 µL, Cemented-Needle (YO-07938-00)
Standard Microliter Syringes, 10 µL, Removable-Needle (YO-07938-10)
Standard Microliter Syringes, 100 µL, Removable-Needle
Standard Microliter Syringes, 250 µL, Removable-Needle.
More Details
2 Ml 13 Mm Vial Clear
Buy 2 Ml 13 Mm Vial Clear Items Including:
Cole-Parmer snap vials; 2 mL, 12 mm X 32 mm, clear
Cole-Parmer snap vials; 2 mL, 12 mm X 32 mm, clear, graduated white m
Cole-Parmer wide neck vials; 2 mL, 12 mm X 32 mm, Clear
Cole-Parmer screw-thread vials; 2 mL, clear, graduated white marking s.
More Details
1/8 Din Panel-Mount
Buy 1/8 Din Panel-Mount Items Including:
1/8 DIN panel-mount programmable tachometer/counter/totalizer, 230 VAC (YO-08212-45)
Temperature Controller, TC, 1/8-DIN, Relay Output, 2 alarms (DC1020CT-102-000-E)
Temperature Controller, TC, 1/8-DIN, Current Output, 2 alarms (DC1020CT-302-000-E).
More Details
Pharmed Bpt
Buy Pharmed Bpt Items Including:
Tubing sets, PharMed BPT with 2 welded stops, 400 mm length, 0.25 mm I (SC0740)
3-Stop Pharmed BPT Tubing, 2.29 mm ID, 12/pack
PharMed BPT Tubing,1/50x9/64",25'
PharMed BPT Tubing,1/32x5/32",25'
PharMed BPT Tubing,1/32X1/8, 25' per pack.
More Details
Specific Gravity Bottles Pycnometer 50ml
Buy Specific Gravity Bottles Pycnometer 50ml Items Including:
Pycnometer, 11.5-mL capacity, 1/8" (3.2-mm) hole in lid (03247Q)
Bottle, Gay-Lussac, with stoppers, unadjusted for calibration in the l (LG-3530-104)
Bottle, Gay-Lussac, with stoppers, adjusted for calibration in the lab (LG-3540-100).
More Details
70618
results
<<<Previous
21
22
23
24
25
26
27
28
29
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Q Tips Swabs to Specific Gravity Bottles Pycnometer 50ml
>
31125329-s-toc_2 31125329stoc2