Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 26

Neoprene Metric Tubing Sizes   to   Big Blue Filter Cartridge

Image
  Product Detail
More Detail

Neoprene Metric Tubing Sizes

Buy Neoprene Metric Tubing Sizes Items Including:
  • Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41)
  • Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-02)
  • Compression Straight Union, Stainless Steel, 6mm OD Tubing (YO-31528-33).

Acu Rite Refrigerator and Freezer Thermometer

Buy Acu Rite Refrigerator and Freezer Thermometer Items Including:
  • Digital-Bottle Refrigerator/Freezer Thermometer with solution filled b
  • Digital-Bottle High Accuracy Refrigerator/Freezer Thermometer with sol
  • Jumbo Refrigerator/freezer thermometer with a 5ml bottle that fits in (4627CP).

Bulkhead Fitting with Barb

Buy Bulkhead Fitting with Barb Items Including:
  • Quick-disconnect fittings, Valved Bulkhead panel mount hose barbs, PP, (HFCD161012)
  • Quick-disconnect fittings, Valved Bulkhead panel mount hose barbs, PP, (HFCD161212)
  • Quick-disconnect fittings, Straight-through Bulkhead panel mount hose (HFC16612).

Ph Test Strips for Water

Buy Ph Test Strips for Water Items Including:
  • EM Quant pH test strips, 0-14 pH
  • Six packs of 100.
  • pHydrion plastic pH indicator strips, 0.0 to 14.0, flip top vial packa
  • pHydrion plastic pH indicator strips, 0.0 to 6.0, flip top vial packag
  • EM Quant® Zinc Test Strips (10038-1).

Npt Luer Adapter

Buy Npt Luer Adapter Items Including:
  • Female luer x 1/8-27 NPT thread, Nylon, 25/pk (YO-45505-78)
  • Female luer x 1/8-27 NPT thread elbow, Nylon, 25/pk (YO-45505-80)
  • Female luer x 1/8-27 NPT thread elbow, PP, 25/pk (YO-45503-80)
  • Male luer lock x 1/8" NPT, 316 SS, each (YO-31507-44).

Digital Fridge Freezer Thermometer

Buy Digital Fridge Freezer Thermometer Items Including:
  • Digital-Bottle Refrigerator/Freezer Thermometer with solution filled b
  • Refrigerator/freezer thermometer with 5ml bottle probe
  • Refrigerator/freezer thermometer with bottle probe (4127CP)
  • Cooper-Atkins Big Digital Display Thermometer, NSF approved.

Calibrated Refrigerator Freezer Thermometer

Buy Calibrated Refrigerator Freezer Thermometer Items Including:
  • Digital-Bottle Refrigerator/Freezer Thermometer with solution filled b
  • Digital-Bottle High Accuracy Refrigerator/Freezer Thermometer with sol
  • Jumbo Refrigerator/freezer thermometer with a 5ml bottle that fits in (4627CP).

Gram Balance Scale

Buy Gram Balance Scale Items Including:
  • Vinyl Dust Cover for Ohaus Cent-O-Gram and Dial-O-Gram balances (01000
  • Ohaus Dial-O-Gram Triple Beam Balance with Tare and Stainless Steel Pa
  • Dial-O-Gram® Triple Beam Balance, 610g X 0.1g, Stainless Steel Pan and.

Roto Torque Rotators

Buy Roto Torque Rotators Items Including:
  • Cole-Parmer Roto-Torque Variable Speed Rotator, 115 VAC, 50/60 Hz
  • Cole-Parmer Roto-Torque Variable Speed Rotator, 230 VAC, 50/60 Hz
  • Test Tube Basket for Cole-Parmer Roto-Torque Variable Speed Rotator
  • Clips 10-15mm Size (YO-07637-41).

Oakton Ph Buffer Solution Msds

Buy Oakton Ph Buffer Solution Msds Items Including:
  • Oakton pH buffer pouches; pH 4.01, 20/pk
  • Oakton pH buffer pouches; pH 7.00, 20/pk
  • Oakton pH buffer pouches; pH 10.00, 20/pk
  • Oakton 4.01 pH buffer, 500-mL
  • Oakton 1.68 pH buffer, 500-mL

Mini Stainless Steel Funnel

Buy Mini Stainless Steel Funnel Items Including:
  • Stainless steel funnel, 2 3/8 oz (YO-07263-00)
  • Stainless steel funnel, 13 oz (YO-07265-00)
  • Funnel rack for modular stainless steel drying racks
  • Justrite Safety Drum Funnel, manual closing, 1" stainless steel flame
  • 150ml Funnel, Kg25 (311208).

Luer Lock Male to Male

Buy Luer Lock Male to Male Items Including:
  • Stopcocks with Luer Connections; 3-way; male lock; 5 flow pattern; 100 (YO-30600-07)
  • Polycarbonate fittings; female luer x male lock, rotating; 10/pack (YO-30600-32)
  • Polycarbonate fittings; female luer x male lock, rotating; 100/pack (YO-30600-33).

Drinking Water Test Strips Copper

Buy Drinking Water Test Strips Copper Items Including:
  • Watersafe Drinking Water Test Kit
  • LaMotte Free Chlorine Test Strips, 0 to 10 ppm
  • LaMotte Total Chlorine Test Strips, 0 to10 ppm
  • LaMotte Total Chlorine Test Strips, 0 to 5 ppm
  • LaMotte Hardness Test Strips, 0 to 180 ppm

Tough-Tags Label Printers and Labels

Buy Tough-Tags Label Printers and Labels Items Including:
  • Small Tough-Tags™ label, clear
  • Small Tough-Tags™ label, white
  • Small Tough-Tags™ label, red
  • Small Tough-Tags™ label, yellow
  • Small Tough-Tags™ label, green

Nalgene Lab

Buy Nalgene Lab Items Including:
  • Thermo Scientific Nalgene lab notebook case bound in a hard waterproof (6301-1000)
  • Thermo Scientific Nalgene lab notebook case bound in a hard waterproof (6301-3000)
  • Thermo Scientific Nalgene lab notebook case bound in a hard waterproof (6301-2000).

Nps Thread Fittings

Buy Nps Thread Fittings Items Including:
  • Compression fittings, Male pipe adapter elbow, Kynar,1/4" OD x 1/8" NP (YO-30805-40)
  • Compression fittings, Male pipe adapter elbow, Kynar,1/4" OD x 3/8" NP (YO-30805-42).

Keck Clamps

Buy Keck Clamps Items Including:
  • Keck Ramp Clamp Tubing Clamps, 4.5 mm, 12/pack
  • Keck Ramp Clamp Tubing Clamps, 10 mm, 12/pack
  • Keck Ramp Clamp Tubing Clamps, 14 mm, 12/pack
  • Keck Glassware Adapter, GL-14 threaded Port or 10 mm OD barbed port, 1
  • Keck Flask/Bulkhead Adapter, 10/pack.

Ptfe Caps

Buy Ptfe Caps Items Including:
  • Threaded Cap, PTFE, 1/8" NPT(F) (F-CA01)
  • Threaded Cap, PTFE, 1/4" NPT(F) (F-CA02)
  • Threaded Cap, PTFE, 1" NPT(F) (F-CA08)
  • Replacement YSI DO membrane cap kit, 1.00 mil PTFE (5906)
  • Chemical bottle pourer, 30-mm cap size (310374-0003).

Labnet Centrifuge Pcr Plate Spinner

Buy Labnet Centrifuge Pcr Plate Spinner Items Including:
  • Compact PCR Plate Centrifuge, 120VAC
  • Compact PCR Plate Centrifuge, 230VAC
  • PCR Support Plate for 17416-00 and -05, PCR strips, tubes and plates/2
  • Individual adapters for Prism™ Microcentrifuge, for 0.2 mL PCR, the.

Pyrex Media Bottle Cap

Buy Pyrex Media Bottle Cap Items Including:
  • Pyrex Brand 1395 media storage bottle w/screw cap, 25 mL
  • Pyrex Brand 1395 media storage bottle w/screw cap, 100 mL
  • Pyrex Brand 1395 media storage bottle w/screw cap, 250 mL
  • Pyrex Brand 1395 media storage bottle w/screw cap, 500 mL.

Ultra Low Profile Magnetic Stir Plates

Buy Ultra Low Profile Magnetic Stir Plates Items Including:
  • Digital MegaMag Genie™ Low Range Stirrer, 120 VAC.

Hach Test Reagents for Water

Buy Hach Test Reagents for Water Items Including:
  • Hach Free Chlorine (USEPA) Test Kit (21055-69)
  • Hach Total Chlorine (USEPA) Test Kit (21056-69)
  • Hach Cyanide Test Kit (24302-00)
  • Hach Fluoride (SPADNS) Reagent
  • Hach Total Iron (USEPA) Test Kit (21057-69)

Vacuum Pumps Theory of Operation Schematics

Buy Vacuum Pumps Theory of Operation Schematics Items Including:
  • Hand-operated vacuum pump, 16 mL/stroke, PVC body
  • Hand-operated vacuum pump (without gauge), 32 mL/stroke, PVC body
  • Hand-operated vacuum/pressure pump, 15 mL/stroke, zinc alloy body
  • Repair kit for vacuum/pressure pump 79301-11.

Solvent Pumps

Buy Solvent Pumps Items Including:
  • PVC Solvent/Hydrocarbon Tubing, 0.19 mm ID, 12/pk (95605-10,3-STOP SOLVA)
  • PVC Solvent/Hydrocarbon Tubing, Two-stop, 0.38 mm ID (116-0533-04)
  • PVC Solvent/Hydrocarbon Tubing, Two-stop, 0.51 mm ID (116-0533-05).

Conductivity Standard Solution

Buy Conductivity Standard Solution Items Including:
  • Thermo Scientific Orion Conductivity Standard 1413 uS, 5 X 60 ml (011007)
  • Thermo Scientific Orion Conductivity Standard 111.9 mS, 5 X 60 ml
  • Oakton conductivity solution, 12,880 µS
  • Cole-Parmer Redi-Stor™ Conductivity Probe Storage Solution, 16 ounc.

Food Grade Rotary Hand Pump

Buy Food Grade Rotary Hand Pump Items Including:
  • Pump, Hand-Operated Rotary Drum Pump, Stainless Steel, 10 GPM, 40" int (219)
  • Rotary drum pump, 8 GPM, Polypropylene (3003T)
  • Rotary drum pump, 6 GPM, Ryton & SS
  • Rotary drum pump, High Volumn 17 GPM, Cast-Iron (3003T) (YO-06510-17).

Hdpe Carboy

Buy Hdpe Carboy Items Including:
  • Baritainer Jerry Can, Natural, HDPE/Quoral 10 L
  • Economical Square Carboy w/handle, 2-1/2 Gal - HDPE, 1/ea (YO-06031-80)
  • Thermo Scientific Nalgene rectangular high-density polyethylene carboy (2211-0020)
  • Carboy Rnd Top-handle HDPE 5gl (16314634-84).

Plastic Pipette Dropper Bulk

Buy Plastic Pipette Dropper Bulk Items Including:
  • Disposable Droppers, LDPE, 1.5 mL, 100/pk (262175)
  • Disposable Droppers, LDPE, 5 mL, 100/pk (262185)
  • Cole-Parmer sterile Pipette, bulk wrapped, plastic blue TD ring, 5mL,
  • Cole-Parmer sterile Pipette, bulk wrapped, plastic orange TD ring, 10m (PB10).

Bev-a-Line Xx Tubing

Buy Bev-a-Line Xx Tubing Items Including:
  • Bev-A-Line XX Tubing, 11/64"ID x 1/4"OD, 500 ft/pack
  • Bev-A-Line XX Tubing, 1/4"ID x 3/8"OD, 500 ft/pack.

Big Blue Filter Cartridge

Buy Big Blue Filter Cartridge Items Including:
  • "Big Blue" industrial filter housing; 10"L, 1-1/2" NPT(F) (150239)
  • "Big Blue" industrial filter housing; 20"L, 3/4" NPT(F) (150468)
  • Fibrillated polypropylene string-wound cartridge, 10" (WPX100BB97P).



Neoprene Metric Tubing Sizes to Big Blue Filter Cartridge

>