Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 27
Nalgene Right
to
Know Safety Bottle
Image
Product Detail
More Detail
Nalgene Right to Know Safety Bottle
Buy Nalgene Right to Know Safety Bottle Items Including:
Thermo Scientific Nalgene "Right-to-Know" safety label customized syst
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for aceto
Thermo Scientific Nalgene "Right-to-Know" safety wash bottle assortmen.
More Details
Pre-Cleaned Bottles
Buy Pre-Cleaned Bottles Items Including:
I-CHEM Certified Pre-Cleaned Wide-Mouth Plastic Bottles, 125 mL
I-CHEM Certified Pre-Cleaned Wide-Mouth Plastic Bottles, 1000 mL
Qorpak Kaptclean™ pre-cleaned sample bottle, 1 liter
Qorpak Precleaned Amber Bottle, 30 mL, PTFE-lined cap, case of 24.
More Details
Modular Stainless Steel Luer Fittings
Buy Modular Stainless Steel Luer Fittings Items Including:
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
Compression Straight Union, Stainless Steel, 12mm OD Tubing (YO-31528-36).
More Details
Flexible Thermocouple Wire
Buy Flexible Thermocouple Wire Items Including:
Type J High Temperature 12"L, .020" diameter thermocouple probe with m (073098A-J-E-12)
Type J High Temperature 12"L, .020" diameter thermocouple probe with m (073098A-J-U-12).
More Details
Digital Wireless Freezer Thermometer
Buy Digital Wireless Freezer Thermometer Items Including:
Digital-Bottle Refrigerator/Freezer Thermometer with solution filled b
Refrigerator/freezer thermometer with 5ml bottle probe
Refrigerator/freezer thermometer with bottle probe (4127CP)
Cooper-Atkins Big Digital Display Thermometer, NSF approved.
More Details
Miniature Hose Barbs
Buy Miniature Hose Barbs Items Including:
Nylon Miniature Check Valve, 1/8" hose barb, fluorosilicone diaphragm (YO-98553-00)
Nylon Miniature Check Valve, 3/16" hose barb, fluorosilicone diaphragm (YO-98553-01)
PVDF Miniature Check Valve, 3/16" hose barb, Viton diaphragm.
More Details
Traceable Refrigerator Freezer Thermometer
Buy Traceable Refrigerator Freezer Thermometer Items Including:
Digital-Bottle Refrigerator/Freezer Thermometer with solution filled b
Digital-Bottle High Accuracy Refrigerator/Freezer Thermometer with sol
Jumbo Refrigerator/freezer thermometer with a 5ml bottle that fits in (4627CP).
More Details
Western Safety Nitrile Blue Gloves
Buy Western Safety Nitrile Blue Gloves Items Including:
Nitrile dipped gloves, nylon liner, M, 12pr (4500-08)
Best N-DEX Nitrile Exam Gloves, Powder-Free, medium, 100/pack
Best N-DEX Nitrile Exam Gloves, Powder-Free, extra-large, 100/pack
Best N-DEX Nitrile Ultimate Gloves, Powder-Free, medium, 50/pack.
More Details
Automatic Zeroing Burettes
Buy Automatic Zeroing Burettes Items Including:
Thermo Scientific Nalgene ® Automatic self-zeroing burette; capacity, (3640-0010)
Thermo Scientific Nalgene ® Automatic self-zeroing burette; capacity, (3640-0025)
Automatic self-zeroing, self-supporting burette; capacity, 10mL; subdi (37918-0109).
More Details
Bottle Vacuum
Buy Bottle Vacuum Items Including:
Corning Storage Bottles; 150-mL volume
Corning Storage Bottles; 250-mL volume
Corning Storage Bottles; 500-mL volume
Whatman™ Klari-Flex™ Filter Bottle; 250 mL Capacity; 24/PK
Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 30 mL, 20 mm Ne.
More Details
Unf to Npt Pipe Fitting
Buy Unf to Npt Pipe Fitting Items Including:
Pipe fittings, Reducing bushing, Nylon, 1/8" NPT x 1/4-28 UNF, 19/32", (YO-31502-21)
Pipe fittings, Nipples UNF, Nylon, 10-32 UNF, 3/8", 1/4", 1/16"; 250/p (YO-31901-44).
More Details
Nalgene Dilution Bottles Polypropylene Copolymer
Buy Nalgene Dilution Bottles Polypropylene Copolymer Items Including:
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-9050)
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0001)
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0002).
More Details
Laboratory Weights
Buy Laboratory Weights Items Including:
High-Molecular Weight Bags, 16 gallon
Scienceware vinyl-covered lead ring weight for 250 to 1000 mL flasks
Scienceware vinyl-covered lead ring weight for 500 to 2000 mL flasks
Scienceware vinyl-covered lead ring weight for 1000 to 4000 mL flasks.
More Details
50 Ml Hdpe Bottle
Buy 50 Ml Hdpe Bottle Items Including:
Thermo Scientific Nalgene Centrifuge bottle; 250-mL; HDPE; 6/pack
Scienceware Plum Covered Sterile Saline (50-mL bottle)/pH Neutralizing
Clear glass dropping bottle, 50 mL
Pack of 6
Inserts for swing-out bucket rotor; 1 50-mL bottle (YO-02550-75).
More Details
Double Wall Tubing
Buy Double Wall Tubing Items Including:
Double-walled PTFE heat-shrink tubing, 0.065"ID, 12 ft/pack (YO-95810-10)
Double-walled PTFE heat-shrink tubing, 0.115"ID, 12 ft/pack (YO-95810-11)
Double-walled PTFE heat-shrink tubing, 0.130"ID, 12 ft/pack (YO-95810-12).
More Details
Stainless Steel Bulkhead Fitting
Buy Stainless Steel Bulkhead Fitting Items Including:
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41).
More Details
Peek Accessories for Liquid Chromatography (Lc)
Buy Peek Accessories for Liquid Chromatography (Lc) Items Including:
PEEK high-pressure unions, tees, and crosses; 0.020" bore, union (YO-02008-02)
PEEK high-pressure unions, tees, and crosses; 0.020" bore, tee (YO-02008-12)
PEEK high-pressure unions, tees and crosses; 0.020" bore, cross (YO-02008-22).
More Details
Inoculating Loops
Buy Inoculating Loops Items Including:
Nichrome V inoculating loop with handle, 3 mm inside diameter, 6" L, n
Nichrome V inoculating loop with handle, 4 mm inside diameter, 8" L, n
Nichrome V inoculating loop with handle, 3 mm inside diameter, 8" L, n.
More Details
32 Mm 5 Micron Syringe Filters
Buy 32 Mm 5 Micron Syringe Filters Items Including:
Whatman 25-mm GD/X Syringe Filters, Polypropylene; pore size, 0.45 m
Cole-Parmer HPLC Syringe Filters, Non Sterile, PTFE, 0.2 um, 30 mm dia
Polycarbonate Filter Holder, 50 mm, 5/pk
Advantec Polypropylene Filter Holder for 25-mm membranes.
More Details
Three Way Stopcock
Buy Three Way Stopcock Items Including:
Thermo Scientific Nalgene PP Three-Way Stopcock, 1/4" to 5/16" hose ba (6470-0002)
PVDF Three-Way Stopcock, 10 mm hose barb, 8 mm orifice
Three-Way Stopcock for Glass and Rigid Tubing, 8 mm tube OD
HDPE Two-Way Stopcock, 10 mm hose barb.
More Details
50 Nm Ptfe Filter Paper
Buy 50 Nm Ptfe Filter Paper Items Including:
Advantec Grade No
101 Qualitative Filter Paper; 12.5 cm dia.; pack of
Advantec Grade No
5C Ashless Quantitative Filter Paper; 12.5 cm dia.;
Advantec PTFE Depth Filter, 10 µm pore, 47-mm diameter; pack of 20.
More Details
Cole-Parmer Tank Brushes
Buy Cole-Parmer Tank Brushes Items Including:
Oval tank brush, polyester bristles, blue, 9"W x 5-1/2" diameter
Oval tank brush, X-shaped nylon bristles, yellow, 9"W x 5-1/2" diamete
Oval tank brush, X-shaped nylon bristles, green, 9"W x 5-1/2" diameter.
More Details
Clear 5 Gallon Water Containers
Buy Clear 5 Gallon Water Containers Items Including:
FDA-approved high-density polyethylene container, 5 gallon (HPB 4716)
Box, HDPE, 5 gallon
Thermo Scientific Orion pH Electrode Storage Solution, 5 gallon (19L)
Bottle, Precleaned, Clear Glass, Square, Wide Mouth, Qorpak, 960 mL, 5.
More Details
70 Micron Nylon Filter
Buy 70 Micron Nylon Filter Items Including:
Corning Disposable Bottletop Filter; 500-mL volume; NYL, membrane; 12/
Gridded Membrane Filters, Non-sterile, White/black, 1.2 Micron
Spectra/Mesh Screen Discs, 55 mm, Nylon, 70 µm, 10/pk
Spectra/Mesh Screen Discs, 47 mm, Nylon, 70 µm, pack of 10.
More Details
Kipp Group Male Luer Lock Fittings
Buy Kipp Group Male Luer Lock Fittings Items Including:
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pac (YO-31200-90)
One-way stopcocks; male luer lock x male luer lock, nickel-plated bras (YO-31507-02).
More Details
British Pipe Thread Fittings
Buy British Pipe Thread Fittings Items Including:
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50)
Threaded adapter, stainless steel, 1/4" female NPT to 1/4" male BSPP (YO-31830-51).
More Details
Microscope Reticle Scale
Buy Microscope Reticle Scale Items Including:
Reticle, Scaled from 10 mm/100 Divisions, 19 mm dia (KR-207-19MM)
Reticle, Scaled from 10 mm/100 Divisions, 21 mm dia (KR-207-21MM)
Reticle, Scaled from 10 mm/100 Divisions, 22.8 mm dia (KR-207-22.8MM)
Eyepiece reticles; Scale 10 mm/0.1mm (MA506).
More Details
Plastic Oval Vial with Snap Lid
Buy Plastic Oval Vial with Snap Lid Items Including:
Thermo Scientific Nalgene Sample vials with snap caps, 12 mL
Snap-cap sample vials, 11.1 mL (19271519-144)
Snap-cap sample vials, 18.5 mL (19272519-144)
Snap-cap sample vials, 25.9 mL (19273619-144)
Snap-cap sample vials, 37.0 mL (19275019-144).
More Details
Reducer to 1 4 Compression Fitting
Buy Reducer to 1 4 Compression Fitting Items Including:
Compression reducing union, Pharmafluor™ PTFE, 1/4" OD x 1/8" OD (YO-06379-11)
Reducing compression union, polypropylene, 5/16 x 1/4" OD, 10 per pack (YO-06381-70)
Reducing Union, 5/16" x 1/4", Nylon, 10 Pack (YO-06382-17).
More Details
Male Luer Lock Syringe
Buy Male Luer Lock Syringe Items Including:
Syringes with PTFE luer lock; syringe capacity; 50µL; needle gauge; (YO-07939-32)
Syringes with PTFE luer lock; syringe capacity; 100µL; needle gauge (YO-07939-33)
Syringes with PTFE luer lock; syringe capacity; 1 mL; needle gauge; 22 (YO-07939-80).
More Details
70618
results
<<<Previous
23
24
25
26
27
28
29
30
31
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Nalgene Right to Know Safety Bottle to Male Luer Lock Syringe
>
31125329-s-toc_2 31125329stoc2