Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 28
Legris Push
to
Connect Tube Fittings
Image
Product Detail
More Detail
Legris Push to Connect Tube Fittings
Buy Legris Push to Connect Tube Fittings Items Including:
Push-to-connect soft tube supports, 3/8" stem x 5/16" tube OD, 10/pack (YO-34007-38)
Push-to-connect soft tube supports, 1/2" stem x 3/8" tube OD, 10/pack (YO-34007-39)
John Guest Push-to-Connect Collet Cover, 5/16" Tube OD.
More Details
Cole-Parmer Hdpe Tote Boxes
Buy Cole-Parmer Hdpe Tote Boxes Items Including:
HDPE tote box, 0.5 cu ft (35-180 BLUE)
HDPE tote box, 1.2 cu ft (35-190 BLUE)
HDPE tote box, 2.6 cu ft (35-230 BLUE)
HDPE tote box, 3.7 cu ft (35-300 BLUE)
Box, HDPE, 2 gallon
More Details
Miniature Electric Vacuum Pumps
Buy Miniature Electric Vacuum Pumps Items Including:
Miniature venturi vacuum pump, 0.5 cfm, 28.0
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 1.5 LPM, 12 VDC, GAS ONLY,
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 1.5 LPM, 24 VDC, GAS ONLY,
Compact/OEM bellows pressure pump, 0.5 LPM 5.8 psi, 3 VDC.
More Details
10 Inch Diameter Stainless Steel Tube
Buy 10 Inch Diameter Stainless Steel Tube Items Including:
160-18 Stainless Steel Pitot Tube 18" Insertion Length 5/16" Diameter
160-48 Stainless Steel Pitot Tube 48" Insertion Length 5/16" Diameter
160-60 Stainless Steel Pitot Tube 60" Insertion Length 5/16" Diameter.
More Details
Glass Jar Supplies
Buy Glass Jar Supplies Items Including:
Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 30 mL, 20 mm Ne
Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 60 mL, 20 mm Ne
Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 120 mL, 22 mm N.
More Details
Ismatec 3 Stop Tubing
Buy Ismatec 3 Stop Tubing Items Including:
Three-stop silicone (peroxide cured) tubing, 0.64 mm ID, 6/pack (SC0106/ 3 STOP)
Three-stop silicone (peroxide cured) tubing, 0.76 mm ID, 6/pack (SCO107/3 STOP)
Three-stop silicone (peroxide cured) tubing, 0.89 mm ID, 6/pack (SC0108/3 STOP).
More Details
Nitex Netting 500 Um Mesh Filter
Buy Nitex Netting 500 Um Mesh Filter Items Including:
Spectra/Mesh Screen Sheets, Polypropylene, 500 µm, 3/pk
Spectra/Mesh Screen Discs, Polypropylene, 90 mm, 500 µm, 10/pk
Spectra/Mesh Screen Discs, Polypropylene, 55 mm, 500 µm, 10/pk.
More Details
Oakton Ph Vs Temperature Tables
Buy Oakton Ph Vs Temperature Tables Items Including:
Electrode pH/Temp double-junction for high pH, low sodium error with 3 (YO-05991-36)
Oakton ATC probe for pH 5, pH 6, Ion 6, pH 11, pH 110, pH 510, Ion 510
Oakton ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 m (YO-35418-05).
More Details
H-B Instruments Hydrometers for Specific Gravity
Buy H-B Instruments Hydrometers for Specific Gravity Items Including:
H-B Instrument Hydrometer, 1.200/1.420Specific Gravity, 0.002 Division
H-B Instrument Hydrometer, 1.000 to 1.225 Specific Gravity, 0.005 Divi
H-B Instrument Hydrometer, 1.200 to 1.450 Specific Gravity, 0.005 Divi.
More Details
Sampling Scoops
Buy Sampling Scoops Items Including:
Scienceware sterile polystyrene sampling scoop, 2 oz
Scienceware sterile polystyrene sampling scoop, 4 oz
Scienceware Earth-Friendly Wide Handle Sampling Spoon, 1.25 ml, 10 pac
Scienceware Earth-Friendly Wide Handle Sampling Spoon, 2.5 ml, 10 pack.
More Details
Cole-Parmer Thermometer
Buy Cole-Parmer Thermometer Items Including:
Cole-Parmer Thermometer/Clock/Humidity Thermometer
Cole-Parmer Extreme-Accuracy Thermometer 25.00°
Cole-Parmer Extreme-Accuracy Thermometer 0.00°, 25.00°, 37.00
Cole-Parmer Big-Digit 4-Alert Alarm Thermometer °C
Cole-Parmer Monitoring Thermometer.
More Details
Dissolved Oxygen Standards
Buy Dissolved Oxygen Standards Items Including:
OptiSphere™ 0.2 NTU Turbidity Standard, 250-mL
OptiSphere™ 0.5 NTU Turbidity Standard, 250-mL
OptiSphere™ 10 NTU Turbidity Standard, 250-mL
OptiSphere™ 200 NTU Turbidity Standard, 250-mL
OptiSphere™ 800 NTU Turbidity Standard, 250-mL.
More Details
Thermo Scientific Ertco Hydrometers
Buy Thermo Scientific Ertco Hydrometers Items Including:
Thermo Scientific ERTCO® Dual-Scale Hydrometer, 1.000 to 2.000 Spe (2545ADS)
Thermo Scientific ERTCO® Dual-Scale Hydrometer, 0.800 to 0.900 Spe (2537DS)
Thermo Scientific ERTCO® Plastic Hydrometer, 0 to 26.5 Percent Sal (2511-02PL).
More Details
Bioprocessing Plastic Tubing Clamps
Buy Bioprocessing Plastic Tubing Clamps Items Including:
Hand tool for plastic hose clamps (YO-06832-97)
Plastic hose clamps, black, 0.569" x 0.650", 100/pack (YO-95613-08)
Screw tubing clamp, 3/bag
Barbed to sanitary adapter, 1/4" ID to 1-1/2" sanitary clamp, animal-f (YO-31513-74).
More Details
Pfa Tube Fittings
Buy Pfa Tube Fittings Items Including:
Pharmafluor Compression To Male NPT Thread Elbow, PFA, 3/8 X 1/4 (YO-31001-52)
Threaded adapter, PFA, 1/8" NPTM x 5/32" tubing ID (YO-06383-90)
Threaded adapter, PFA, 1/8" NPTM x 1/4" tubing ID (YO-06383-91).
More Details
Baume Hydrometer
Buy Baume Hydrometer Items Including:
Durac Baume Polycarbonate Hydrometer, 19 to 31, 0.2 Div, 255mm L, Indi
H-B Instrument Hydrometer, —1 to 12° Baume, 0.1° Divisi
H-B Instrument Hydrometer, —49 to 61° Baume, 0.1° Divis
H-B Instrument Hydrometer, —0 to 75° Baume, 0.5° Divisi.
More Details
Low Vacuum Gauge
Buy Low Vacuum Gauge Items Including:
Ashcroft Dual-Scale Low-Vacuum Gauge, 15" H2O to 0 and 0 to
Ashcroft Dual-Scale Low-Vacuum Gauge, 30" H2O to 0 and 0 to
Ashcroft Dual-Scale Low-Vacuum Gauge, 60" H2O to 0 and 0 to.
More Details
Colder Products
Buy Colder Products Items Including:
Panel Mount Valved Coupling Body for 1/4-28 Port, PP (PMCD18042812)
Male AseptiQuik Connector, 5/pk (YO-20002-88)
Coupling Insert, Polysulfone, 3/4" ID Tube, 10/pk (MPU221239M)
Aseptiquik Standard Insert, 1/2" Hose Barb, High Temperature.
More Details
Cole-Parmer Safety Products
Buy Cole-Parmer Safety Products Items Including:
Safety Sign, Danger Lockout Equipment Before Entering, 7 X 10, Aluminu (MLKT106VA)
Safety Sign, Danger Lockout Equipment Before Servicing, 7 X 10, Adhesi (MLKT007VS)
Safety Sign, Danger Lockout Equipment Before Servicing, 7 X 10, Plasti (MLKT007VP).
More Details
Bausch & Lomb Handheld Magnifiers
Buy Bausch & Lomb Handheld Magnifiers Items Including:
Bausch & Lomb Handheld LED Magnifier, 2" x 4", 2x magnification
Bausch & Lomb Handheld LED Magnifier, 3-1/2" diameter, 2x magnificatio
Measuring Scales;" scale, 0.75" in 0.005" increments (81-34-37).
More Details
Cole-Parmer Microbore Ptfe Tubing
Buy Cole-Parmer Microbore Ptfe Tubing Items Including:
Microbore PTFE Tubing Assortment Pack
Ultramicrobore PTFE Tubing, 0.002"ID 0.014"OD, 100 ft/pack
Ultramicrobore PTFE Tubing, 0.004"ID 0.016"OD, 100 ft/pack
Ultramicrobore PTFE Tubing, 0.006"ID 0.016"OD, 100 ft/pack.
More Details
Laboratory Glass Incubator Thermometers
Buy Laboratory Glass Incubator Thermometers Items Including:
Frio-temp Incubator Thermometer, 10/45c, 98mm Length, Bottle Filled Wi
Frio-temp Incubator Thermometer, 15/50°C, 135mm Length, Bottle Fil (20720T)
Frio-temp Incubator Thermometer, 15/50°C, 135mm Length, Bottle Fil (30720).
More Details
Luer Lock Connector Dimensions
Buy Luer Lock Connector Dimensions Items Including:
Male luer lock plug, PC, 25/pk (YO-45504-56)
Male Luer Lock Plug, PP, 100 Per Pack (YO-45603-56)
Polycarbonate fittings; male lock x male lock, rotating; 10/pack (YO-30600-50)
Male luer with lock ring x female luer coupler, PC, 25/pk (YO-45501-80).
More Details
Ika Mini Color Squid Stirrer
Buy Ika Mini Color Squid Stirrer Items Including:
Ika Digital Color Squid (white) 110-240V
Ika Digital Color Squid (US flag) 110-240V.
More Details
Flo Et Flow Control
Buy Flo Et Flow Control Items Including:
Stainless steel flow sensors with 0 to 5 VDC output; 1 to 10 L/min flo.
More Details
Techne Bath & Circulator Accessories
Buy Techne Bath & Circulator Accessories Items Including:
Polypropylene balls, 10 mm, pack of 5000 (YO-06821-12)
Polypropylene balls, 20 mm, pack of 500 (YO-06821-22)
Polypropylene balls, 20 mm, pack of 2000 (YO-06821-25).
More Details
Upchurch Hplc Valves and Filters
Buy Upchurch Hplc Valves and Filters Items Including:
Four-port switching valve (YO-02012-00)
In-line solvent filter, 2 µm SS frit with PEEK ring (YO-02010-00)
Precolumn filter, 316 SS and PEEK filter, 2 µm frit (YO-02011-00)
Replaceable solvent inlet filter assembly, 2 µ, 10 mL/min max
flow (A-551).
More Details
Nylon Tubing 1 4 Purple Dot
Buy Nylon Tubing 1 4 Purple Dot Items Including:
Barbed Fittings; Elbow Connector, Nylon, 1 1/4" Tubing ID, 5/pk (YO-31408-05)
Barbed Fittings; T Connector, Nylon, 1 1/4" Tubing ID, 5/pk (YO-31408-08)
Barbed Fittings; Straight Connector, Nylon, 1 1/4" Tubing ID, 50/pk (YO-31418-00).
More Details
Eppendorf Microcentrifuge Tube Racks
Buy Eppendorf Microcentrifuge Tube Racks Items Including:
Eppendorf Microcentrifuge Tubes; capacity, 1.5-mL; 500/pack
Reversible 96-well microcentrifuge tube rack, blue
Thermo Scientific Nalgene microcentrifuge tube rack, white
Reversible 96-well microcentrifuge tube rack, assorted colors (5322-39-22CP1).
More Details
Buret Clamp
Buy Buret Clamp Items Including:
Buret Clamp, Single, Flat, Medium
Buret Clamp, Single, Three-prong, Medium
Double Buret Clamp/Stand (916187)
Double Buret Clamp Stainless Steel
Buret Support Stand, Porcelein Stand with Stainless Steel Support Rod
More Details
70618
results
<<<Previous
24
25
26
27
28
29
30
31
32
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Legris Push to Connect Tube Fittings to Buret Clamp
>
31125329-s-toc_2 31125329stoc2