Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 3
Deadweight Testers
to
Water Conductivity Meter
Image
Product Detail
More Detail
Deadweight Testers
Buy Deadweight Testers Items Including:
Ashcroft Deadweight Tester Kit, 1000 psi Maximum
Ashcroft Deadweight Tester Kit, 2000 psi Maximum
Ashcroft Deadweight Tester Kit, 3000 psi Maximum
Ashcroft Deadweight Tester Kit, 5000 psi Maximum
Ashcroft Deadweight Tester Kit, 10,000 psi Maximum.
More Details
Small Rotary Vane Pump
Buy Small Rotary Vane Pump Items Including:
High-pressure, brass-bodied rotary vane pump, 2.3 GPM, 115/230 VAC, 5
High-pressure, 303 stainless steel rotary vane pump, 2.3 GPM, 115/230
High-pressure, 303 stainless steel rotary vane pump, 4.3 GPM, 115/230.
More Details
Cole-Parmer Continuous-Cycle Four-Syringe Pumps
Buy Cole-Parmer Continuous-Cycle Four-Syringe Pumps Items Including:
Cole-Parmer Continuous Flow Syringe Pump, 115V
Cole-Parmer Contniuous Flow Syringe Pump, 115V, Programmable
Cole-Parmer Syringe Pump Accessories, Autofill Low Pressure PPS Pinch
Cole-Parmer Syringe Pump Accessories, Continuous Medium Pressure PTFE.
More Details
Ro Tap Sieve Shaker Manual
Buy Ro Tap Sieve Shaker Manual Items Including:
W.S
Tyler Test Stand for Ro-Tap Shakers, 20
W.S
Tyler Wet Test Kit for 8" Diameter Ro-tap Shakers Only
W.S
More Details
Lightnin Mixer
Buy Lightnin Mixer Items Including:
Lightnin LabMaster Analog Remote Control Mixer with Tube Suspension Mo
Lightnin LabMaster Analog Remote Control Mixer with Universal Clamp Mo
Lightnin® 316 SS high-shear radial flow impeller (Lightnin® R- (YO-04560-30).
More Details
Corning Ph Meter Model no 430
Buy Corning Ph Meter Model no 430 Items Including:
Replacement pH electrode, waterproof, for meter kits 58901-00 and -05
pH electrode refillable single-junction for Corning meters (YO-27507-28)
YSI splashproof dissolved oxygen meter with probe, 25-ft cable, model
YSI Model 556 Multiparameter Meter.
More Details
High Viscosity Transfer Pumps
Buy High Viscosity Transfer Pumps Items Including:
High-viscosity tube set (F55GS50/2150/21-39)
High-Viscosity, 4 to 20 mA Pace Pump, 12 GPD, 115 VAC, 50/60 Hz
High-Viscosity, 4 to 20 mA Pace Pump, 240 GPD, 115 VAC, 50/60 Hz
High-Viscosity, 4 to 20 mA Pace Pump, 240 GPD, 230 VAC, 50/60 Hz.
More Details
Lindberg Blue Furnace
Buy Lindberg Blue Furnace Items Including:
Ceramic Hearth Plate for Thermo Scientific Lindberg Blue M Muffle Furn (7011-2022-00B)
Single Setpoint Controller for Lindberg Blue M™ Split Hinge Tube Fu (CC58114A-1)
Single Setpoint Controller for Lindberg Blue M™ Split Hinge Tube Fu (CC58114C-1).
More Details
Heated Stainless Steel Funnel
Buy Heated Stainless Steel Funnel Items Including:
SAHARA Heated Stainless Steel Circulating Bath, SC-100 controller, 2-5 (152-1031)
SAHARA Heated Stainless Steel Circulating Bath, SC-100 controller, 4-7 (152-1078)
SAHARA Heated Stainless Steel Circulating Bath, SC-100 controller, 4-7 (152-1071).
More Details
Universal Centrifuge
Buy Universal Centrifuge Items Including:
High-Speed Programmable Refrigerated Universal Centrifuge; 120V (YO-17303-10)
Compact Variable-Speed Universal Centrifuge Tube Adapter for 15-mL Fix (YO-17306-60)
Compact Variable-Speed Universal Centrifuge Tube Adapter for 50-mL Fix (YO-17306-64).
More Details
Ultra-Quiet Air Compressors
Buy Ultra-Quiet Air Compressors Items Including:
Ultra-Quiet Oil-lubricated Air Compressor, 0.5 cfm, 115 VAC (P15-TC)
Ultra-Quiet Oil-lubricated Air Compressor, 1.5 cfm, 115 VAC (P50/24AL)
Ultra-Quiet Oil-lubricated Air Compressor, 4.5 cfm, 115 VAC (P150/50AL).
More Details
Water Activity Meters
Buy Water Activity Meters Items Including:
LabSwift Portable Water Activity Instrument
+GF+ Signet I/O Module, 2 inputs, 2 active 4 to 20 mA outputs (3-8900.401-3)
Activate™ High-Level Chlorine Test Strips 50/PK (77085)
eVALC Filter for Low Alcohol, for use with LabTouch and LabMaster Wate.
More Details
Orbital and Reciprocating Shakers
Buy Orbital and Reciprocating Shakers Items Including:
Lab
Companion Dual-Action Shaker, 14" x 14", 120V
Lab
Companion Dual-Action Shaker, 14" x 14", 240V
Lab
More Details
Titration Equipment
Buy Titration Equipment Items Including:
Aquamax Volumetric Karl Fischer Titrator (YO-94100-01)
Aquamax Total Acid Number (TAN) Titrator
Aquamax Coulometric Karl Fischer Titrator
Schott® manual volumetric titrator, 20-mL capacity, 230 VAC (YO-24906-05)
Schott universal titrator, 115 VAC.
More Details
Mechanical Stirrers
Buy Mechanical Stirrers Items Including:
Cole-Parmer Five-Position Stirrer, 120 VAC
Multi-position Stirrer, 5 position 100-240V
Multi-position Stirrer, 10-position 100-240V
Multi-position Stirrer, 15 position 100-240V
Cole-Parmer economical mechanical convection oven, 1.7 cu ft, 120 VAC.
More Details
Atago Handheld Manual Refractometers
Buy Atago Handheld Manual Refractometers Items Including:
Atago Three-range sugar refractometer, 0 to 90% Brix
Atago Brix Refractometer, 28.0 to 62.0% Brix w/automatic temperature c
Atago Brix Refractometer, 45.0% to 82.0% Brix, with Automatic Temperat
Atago Brix Refractometer, 45.0% to 82.0% Brix.
More Details
Cole-Parmer Masterflex Pump
Buy Cole-Parmer Masterflex Pump Items Including:
Cole-Parmer Touch-Screen Peristaltic Dispensing Pump with Masterflex L (YO-07582-00)
Piston Pump System consisting of Masterflex L/S Drive, adapter and Q2 (7752140, 0710448, 0710466)
Handheld Remote Controller; DB15(M) Connector; 6 ft (1.8 m) Cable.
More Details
Air Cadet Vacuum Pump
Buy Air Cadet Vacuum Pump Items Including:
Air Cadet Vacuum/Pressure Pump, OEM, 0.65 CFM, 20" HG, 115 VAC
Air Cadet Vacuum/Pressure Pump, Diaphragm, single head, 0.5 cfm, 230 V
Air Cadet PRO, 0.6 CFM, Standard duty, 230 VAC
Replacement Valox pump head assembly for all air Cadet pumps.
More Details
Thermolyne Furnace
Buy Thermolyne Furnace Items Including:
Thermo Scientific Thermolyne Atmosphere-Controlled Programmable Muffle (F6028C-60-80)
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in ch (F47915)
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in ch (F47910-33).
More Details
Calibration Vacuum Pumps
Buy Calibration Vacuum Pumps Items Including:
Pneumatic Pressure and Vacuum Pump, 0-600 PSI; 0-28.5" Hg
Low-Pressure and Vacuum Hand Pump, 0-45 PSI
Pump/Vacuum Kit 60
Pneumatic pump & vacuum, 600 psi to -28 in Hg
Additel 936 Bench-top Hydraulic Calibration Pump, 85% Vacuum to 15,000.
More Details
Nanopure Water System Splash Guards
Buy Nanopure Water System Splash Guards Items Including:
Nanopure Life Science Systems (RO/Distilled Feed) Consumables Bundle (D502144)
Thermo Scientific Barnstead Replacement Ultrafilter for NANOpure™ R (FL550X1)
Organic-Free RO or Distilled Feed Cartridge Pack for Thermo Scientific (D50280).
More Details
Oakton Meters Wd 35801 00
Buy Oakton Meters Wd 35801 00 Items Including:
Oakton pH electrode, epoxy body, sealed, single-junction, general purp (YO-35801-00)
Oakton deionized water rinse solution, box of 20 pouches.
Oakton Waterproof PCD 650 Multiparameter Meter and Probes
Oakton Waterproof pH/Con 300 Meter with probes.
More Details
Knf Vacuum Pumps
Buy Knf Vacuum Pumps Items Including:
Explosion-proof vacuum pump, 1.2 cfm, 27" Hg, 115 VAC (N035 STI-EX)
Pump, Vacuum Pump with Explosion Proof Motor, Vacuum and Pressure, Alu (YO-07057-84)
Pump, Vacuum Pump with Explosion Proof Motor, Vacuum only, Aluminum he (YO-07057-85).
More Details
Brinkmann Laboratory Equipment
Buy Brinkmann Laboratory Equipment Items Including:
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog 0.7 cu ft, 12 (23210100)
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog, 0.7 cu ft, 2 (23210151)
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog 0.8 cu ft, 12 (23210304).
More Details
Cole-Parmer Environmental Chambers
Buy Cole-Parmer Environmental Chambers Items Including:
Controlled Environmental Chamber, 115 VAC, 60z (5518-115V)
Controlled Environmental Chamber, 230 VAC, 50 Hz (5518-220V)
Controlled Humidity Chamber, 115 VAC (5506-115V)
Controlled Humidity Chamber, 230 VAC (5506-220V)
Mini Glove Chamber (5503-11).
More Details
Masterflex Tubing Size
Buy Masterflex Tubing Size Items Including:
Masterflex B/T Peroxide-Cured Silicone Tubing, Size 87, 3 ft (YO-96406-87)
Masterflex B/T BioPharm Plus Platinum-cured Silicone Tubing, Size 91, (YO-96444-91)
Gore High-Resilience Style 400 Pump Masterflex Precision Tubing, size (YO-06439-16).
More Details
Industrial Large Stainless Steel Funnel
Buy Industrial Large Stainless Steel Funnel Items Including:
Stainless steel funnel, 2 3/8 oz (YO-07263-00)
Stainless steel funnel, 6 3/8 oz (YO-07264-00)
Stainless steel funnel, 13 oz (YO-07265-00)
Stainless steel funnel, 22 1/4 oz (YO-07266-00)
Stainless steel funnel, 32 oz (YO-07267-00).
More Details
Variable Speed Mixer from Arrow 850
Buy Variable Speed Mixer from Arrow 850 Items Including:
Light-Duty, Dual-Shaft Variable Speed Mixer, 120 VAC (YO-04330-00)
Heavy-Duty, Variable Speed Mixer, 120 VAC (YO-04340-10)
Waring variable-speed blender, 1-liter, SS, 120v
Lightnin high-torque direct-drive mixer, variable-speed, fits 2" NPT b.
More Details
Knf Pumps
Buy Knf Pumps Items Including:
Explosion-proof vacuum pump, 1.2 cfm, 27" Hg, 115 VAC (N035 STI-EX)
Pump, Vacuum Pump with Explosion Proof Motor, Vacuum and Pressure, Alu (YO-07057-84)
Pump, Vacuum Pump with Explosion Proof Motor, Vacuum only, Aluminum he (YO-07057-85).
More Details
Water Conductivity Meter
Buy Water Conductivity Meter Items Including:
Cole-Parmer Digital, Field Conductivity Meter
accumet® AB 30 benchtop conductivity meter kit
accumet® XL 30 benchtop conductivity meter only
Horiba E-51 Waterproof Conductivity Meter
Cole-Parmer Traceable Portable Conductivity Meter (YO-19601-03).
More Details
70618
results
<<<Previous
1
2
3
4
5
6
7
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Deadweight Testers to Water Conductivity Meter
>
31125329-s-toc_2 31125329stoc2