Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 30
Liebert Float Switch Ls 7
to
Pyrex Brand 2103 Class a Burettes
Image
Product Detail
More Detail
Liebert Float Switch Ls 7
Buy Liebert Float Switch Ls 7 Items Including:
Standard float switch; polypropylene (solid), 3/8"-16 thread mounting (171514)
Standard float switch; stainless steel with slosh shield, (2) 9/32" mo (43765)
LS-3 Level Switch, Polypropylene Float 0.60 Minimum Liquid Specific Gr (171517).
More Details
Extra Long Latex Gloves
Buy Extra Long Latex Gloves Items Including:
Microflex Evolution One Latex Gloves; size, extra-large
Microflex Diamond Grip™ Latex Gloves; size, extra-large
Microflex Diamond Grip Plus™ Latex Gloves; size, extra-large
Best HD™ Liquid-Proof Latex Gloves, Rolled Cuff, extra-large, 15"L,.
More Details
Digital Timer Clock
Buy Digital Timer Clock Items Including:
Extech Digital Stopwatch/Clock
Cole-Parmer triple-display clock/timer, NIST-traceable calibration rep
Cole-Parmer eight-channel clock/timer with adjustable volume
Triple-display clock/timer (YO-94440-00)
Cole-Parmer Color Frame Four-Channel Timer.
More Details
Tubing Unions
Buy Tubing Unions Items Including:
Compression reducing union, Pharmafluor™ PTFE, 1/4" OD x 1/8" OD (YO-06379-11)
Straight compression union, Pharmafluor™ PTFE, 1/4" tubing OD (YO-06396-21)
PTFE Valves, Micro Tubing Valves, 2-way, 1 To 4 mm OD, 2/pack (YO-06473-10).
More Details
Cardboard Microcentrifuge Tube Racks
Buy Cardboard Microcentrifuge Tube Racks Items Including:
Thermo Scientific Nalgene microcentrifuge tube rack, white
Cover for 96-well microcentrifuge tube racks
Microcentrifuge Tube Rack (holds 50 X 1.5 mL Tubes) (H18905-0001)
Reversible 96-well microcentrifuge tube rack, assorted colors (5322-39-22CP1).
More Details
Forceps Types
Buy Forceps Types Items Including:
Steel forceps with PTFE coating (37937)
Heavy Duty PTFE Forceps Square End 7 7/8" (316504-0002)
Heavy Duty PTFE Forceps Sharp End 4" (316514-0000)
Disposable forceps, reinforced nylon (DF8088N)
Disposable sterile polystyrene forceps, PS.
More Details
Male to Male Luer Lock Adapter
Buy Male to Male Luer Lock Adapter Items Including:
Luer fittings, 316 SS fittings, male luer lock x 1/8" hose barb (YO-31507-25)
Luer fittings, 316 SS fittings, male luer lock x 1/16" hose barb (YO-31507-26)
Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62).
More Details
Nylon Threaded Fittings
Buy Nylon Threaded Fittings Items Including:
Barbed fitting kits, Threaded barb adapter kits,Black Nylon, small fit (YO-31514-10)
Barbed fitting kits, Threaded barb adapter kits, Nylon, large fittings (YO-31514-34)
Straight threaded coupling, nylon, 1/8" NPT(F), pack of 6 (YO-06349-00).
More Details
Polypropylene Pipe Fittings Threaded
Buy Polypropylene Pipe Fittings Threaded Items Including:
Pipe fittings, Nipple, Natural PP, 1/8" NPT, 1-3/32", 17/32", 1/4"; 10 (YO-40610-90)
Pipe fittings, Nipple, Natural PP, 1/4" NPT, 1-1/2", 21/32", 11/32"; 1 (YO-40610-91).
More Details
Test Tube Racks in Atlanta
Buy Test Tube Racks in Atlanta Items Including:
Scienceware test tube rack for 10- to 13-mm tubes
Scienceware polypropylene test tube rack for 26-30 mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, w
Thermo Scientific Nalgene Unwire™ test tube rack for 13-mm tubes, o.
More Details
Gloves Best Viton 890
Buy Gloves Best Viton 890 Items Including:
Best Viton Gloves, extra-large, 28-mil thickness, 1 pair.
More Details
Navy Cotton Lab Coats with Snaps
Buy Navy Cotton Lab Coats with Snaps Items Including:
Unisex Poly-Cotton Lab Coat, Navy, XXX-Large (54-56) (6116-14-SZ XX)
Unisex Poly-Cotton Lab Coat, White, XX-Large (50-52) (6116-11-SZ X)
Unisex Poly-Cotton Lab Coat, White, XXX-Large (54-56) (6116-11-SZ XX).
More Details
Ldpe Tubing
Buy Ldpe Tubing Items Including:
Low-Density Polyethylene (LDPE) Tubing, 5/64" x 1/8", 100 ft/bag
Low-Density Polyethylene (LDPE) Tubing, 7/64" x 5/32", 100 ft/bag
Polyethylene Tubing, 0.118x3/16, 100' per pack
Polyethylene Tubing, 0.106x5/32, 100' per pack.
More Details
Ultra Low Temperature Thermometer
Buy Ultra Low Temperature Thermometer Items Including:
FRIO-TEMP ULTRA LOW FREEZER THERMOMETER, -130/80F 145MM LENGTH, BOTTLE
DURAC , Bule Spirit, Verification Thermometer, -90/25C, Ultra Low Free
Frio-temp Certified Verification Thermometer For Ultra Low Freezers, P.
More Details
Female Luer Lok Syringe
Buy Female Luer Lok Syringe Items Including:
BD™ bulk syringes, non-sterile clean, 3mL Luer-Lok™ tip, 1600/ca (YO-07945-02)
BD™ bulk syringes, non-sterile clean, 5mL Luer-Lok™ tip, 1400/ca (YO-07945-06)
BD™ bulk syringes, non-sterile clean, 10mL Luer-Lok™ tip, 850/ca (YO-07945-12).
More Details
H2o Vacuum Gauge
Buy H2o Vacuum Gauge Items Including:
Ashcroft Dual-Scale Low-Vacuum Gauge, 15" H2O to 0 and 0 to
Ashcroft Dual-Scale Low-Vacuum Gauge, 30" H2O to 0 and 0 to
Ashcroft Dual-Scale Low-Vacuum Gauge, 60" H2O to 0 and 0 to.
More Details
Tri Clover Screen Gasket Filters
Buy Tri Clover Screen Gasket Filters Items Including:
Silicone Gasket, 10 MESH, Size 1-1/2" Tri-Clamp (YO-30523-06)
Silicone Gasket, 20 MESH, Size 1/2" Tri-Clamp (YO-30523-20)
Silicone Gasket, 20 MESH, Size 1-1/2" Tri-Clamp (YO-30523-26).
More Details
Replacement Microscope Slide Clips
Buy Replacement Microscope Slide Clips Items Including:
Superfrost Excell Clipped Corner Adhesion Microscope Slides, 144/pk
Plain Clipped-Corner Slides, 144/PK (2950-4-001)
Frosted Clipped-Corner Slides, 144/PK (2951-4-001)
Superfrost Clipped-Corner Slides, 144/PK (4951-4-001).
More Details
Cosmetic Sample Jars 1 4 Oz
Buy Cosmetic Sample Jars 1 4 Oz Items Including:
Kimble Straight-Sided Jar, Amber, 4 oz, 24/cs
Hinged-Lid Sample Containers, PP, 1/4 oz, 2500/pk.
More Details
Cga 180
Buy Cga 180 Items Including:
Single-Stage Regulator, 180 Capacity, 350 CGA Fitting (YO-98200-02)
Single-Stage Regulator, 180 Capacity, 580 CGA Fitting (YO-98200-12).
More Details
Female to Female Luer Lock Adapter
Buy Female to Female Luer Lock Adapter Items Including:
adapter; female luer lock to two-way valve; Kynar body and PP insert.
More Details
Wide-Mouth Bottles to 1000 Ml
Buy Wide-Mouth Bottles to 1000 Ml Items Including:
Thermo Scientific Nalgene low-density polyethylene wide-mouth bottle, (2103-0032)
Thermo Scientific Nalgene low-density polyethylene wide-mouth bottle, (2103-0001)
Thermo Scientific Nalgene Wide-Mouth Economy Bottle, PPCO, 1000 mL, 6/ (YO-06034-71).
More Details
Restaurant Heat Resistant Rubber Gloves
Buy Restaurant Heat Resistant Rubber Gloves Items Including:
Ansell Thermaprene™ Heat-Resistant, Lined Neoprene Gloves, size 10,
MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Medium
MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Extra-large
Best D-Flex™ Cut-Resistant Glove, Full-Face Natural Rubber Dotted C.
More Details
Laboratory Spatulas
Buy Laboratory Spatulas Items Including:
Nickel/stainless steel laboratory spatula, with 1 5/8"L flat rounded e (3100)
Nickel/stainless steel laboratory spatula, with 1 5/8"L flat tapered e (3090)
Nickel/stainless steel laboratory spatula, with 1 1/2"L flat tapered e (3080).
More Details
Chlorine Dioxide Indicator Strips
Buy Chlorine Dioxide Indicator Strips Items Including:
Test Kit, Chlorine Dioxide, 0-10 ppm (3622)
Test Strips, Chlorine Dioxide, 0-10ppm, 50 strips/vial (2999)
Test Strips, Chlorine Dioxide, 0-500ppm, 50 strips/vial (3002).
More Details
Female Luer T Connector
Buy Female Luer T Connector Items Including:
Female luer x female luer adapter, PP, 25/pk (YO-45500-22)
Female luer cap with tether loop, PP, 25/pk (YO-45500-64)
Male luer with lock ring x female luer coupler, PP, 25/pk (YO-45500-80)
Female luer x female luer adapter, PC, 25/pk (YO-45501-22).
More Details
Small Diameter Steel Tubing
Buy Small Diameter Steel Tubing Items Including:
Type J Small-diameter thermocouple probe with miniature stainless stee (072997E-J)
Type K Small-diameter thermocouple probe with miniature stainless stee (072997E-K)
Type T Small-diameter thermocouple probe with miniature stainless stee (072997E-T).
More Details
Cpc Plastic Fitting Panel Mount
Buy Cpc Plastic Fitting Panel Mount Items Including:
Quick-disconnect fittings, Panel-mount John Guest straight-through fit (PLC11004)
Quick-disconnect fittings, Panel-mount John Guest valved fittingDelrin (PLCD41004)
Quick-disconnect fittings, Panel-mount John Guest valved fittingDelrin (PLCD11004).
More Details
Beaker Types
Buy Beaker Types Items Including:
Polypropylene graduated low-form beaker with handle, 1000 mL
Beaker w/handle PP 2000ml 2/pk
Beaker w/handle PP 3000ml 2/pk
Borosil, Beaker, 100ml/12pk (BG1060-100)
Borosil, Beaker, 500ml/6pk (BG1060-500)
More Details
Pyrex Brand 2103 Class a Burettes
Buy Pyrex Brand 2103 Class a Burettes Items Including:
PYREX Brand 2103 Burette; capacity, 10 mL; graduation interval, 0.05mL
PYREX Brand 2103 Burette; capacity, 25 mL; graduation interval, 0.10mL
PYREX Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL.
More Details
70618
results
<<<Previous
26
27
28
29
30
31
32
33
34
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
44
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Liebert Float Switch Ls 7 to Pyrex Brand 2103 Class a Burettes
>
31125329-s-toc_2 31125329stoc2