Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 300
Two Way Radios Truck Unit
to
Thick Frosted Glass Cylinders Candle Covers
Image
Product Detail
More Detail
Two Way Radios Truck Unit
Buy Two Way Radios Truck Unit Items Including:
CLS Two-Way Radio, UHF, 1 watt, 1 channel
CLS Two-Way Radio, UHF, 1 watt, 4 channel
Single-Channel VHF radio
Six-channel UHF radio.
More Details
Stainless Steel Lab Fittings
Buy Stainless Steel Lab Fittings Items Including:
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41).
More Details
Outdoor Thermometers Metal
Buy Outdoor Thermometers Metal Items Including:
12" Dial Metal Thermometer -60F to 120F and -50C to 50C
Suitable for (49562J)
12" Vertical Metal Wall Thermometer with Alcohol Spirit Tube
-50F to
12" Dial Metal Thermometer -60F to 120F and -50C to 50C
More Details
Disposable Rotor-Stator Generator
Buy Disposable Rotor-Stator Generator Items Including:
Rotor-Stator Generator, 7-mm diameter, 95 mm, fine; sample volume rang (YO-04727-68)
Rotor-Stator Generator, 7-mm diameter, 95 mm, course; sample volume ra (YO-04727-70).
More Details
120 Ohm Platinum Rtd
Buy 120 Ohm Platinum Rtd Items Including:
Cole-Parmer 100 ohm Platinum RTD Probes; 5"L
Cole-Parmer 100 ohm Platinum RTD Probes; 9"L
GLI 1000 Ohm electrodeless conductivity sensor, PEEK, 3/4 (3727E2T).
More Details
3 Point Vacuum Pump
Buy 3 Point Vacuum Pump Items Including:
Cole-Parmer® Two Stage Rotary Vane Vacuum Pump, 106/128L/min, 3x10 (YO-72226-65)
Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698123)
Vacuubrandreg Rotary Vane Vacuum Pump, 1.6 cfm, 1.5 X 10-3/sup (698120).
More Details
Large Nalgene Liquid Dispensers
Buy Large Nalgene Liquid Dispensers Items Including:
Thermo Scientific Nalgene Filling bell for liquid transfer, 4-1/2"H x.
More Details
Adjustable Hydraulic Check Valves Knurl
Buy Adjustable Hydraulic Check Valves Knurl Items Including:
PV 623 hydraulic presure relief valve, adjustable in the range 10 to 5 (IO620-PRV-H-1)
PV 623 hydraulic presure relief valve, adjustable range 50 to 200 bar/ (IO620-PRV-H-2).
More Details
Cleanroom Biohazard Spill Kit
Buy Cleanroom Biohazard Spill Kit Items Including:
Lab & Cleanroom Poly Drum Spill Station.
More Details
36 Tube
Buy 36 Tube Items Including:
Masterflex Standard pump head for L/S 36 tubing, PC housing/SS rotor.
Foxy R1 Fraction Collector with 1 Rack; rack holds 36 tubes with 25mm (YO-01466-40)
Assembly Of Flared 0.062 X 0.125" PTFE Tubing, 36", With 1/4-28 Click- (YO-96154-36).
More Details
Kimtech Pure G3
Buy Kimtech Pure G3 Items Including:
KIMTECH Pure G3 NXT Nitrile Cleanroom Gloves, Size 7, 60/bag
KIMTECH Pure G3 NXT Nitrile Cleanroom Gloves, Size 7.5, 60/bag
KIMTECH Pure G3 NXT Nitrile Cleanroom Gloves, Size 8, 60/bag
KIMTECH Pure G3 NXT Nitrile Cleanroom Gloves, Size 8.5, 60/bag.
More Details
Drug Compounding Equipment Supplies
Buy Drug Compounding Equipment Supplies Items Including:
Buffing compound remover for ultrasonic cleaners, 1 gallon bottle (000-955-316).
More Details
Fischer Titrators
Buy Fischer Titrators Items Including:
Aquamax Coulometric Karl Fischer Titrator
Aquamax Volumetric Karl Fischer Titrator (YO-94100-01).
More Details
Hubtron Tubing
Buy Hubtron Tubing Items Including:
Tubing, Hubtron™ Basic, 1/16" x 3/16", 25' (YO-95225-00)
Tubing, Hubtron™ Basic, 1/8" x 1/4", 25' (YO-95225-02)
Hubtron Black Pascal Tubing, 5/16 x 7/16, 25 ft/pk (YO-95257-08).
More Details
Peristaltic Pump Tubing Adapter
Buy Peristaltic Pump Tubing Adapter Items Including:
L/S NEMA type 56C pump head adapter; 3.7:1 adapter gear ratio; 2220 ma
Masterflex two-stop pump tubing, Tygon, L/S 15
Pack of 4.
Masterflex two-stop pump tubing, Tygon, L/S 24
Pack of 4.
More Details
Small Glass Bottles Vials with Tops
Buy Small Glass Bottles Vials with Tops Items Including:
Rack insert for BOD bottles; inserts on either top or bottom rack, hal (YO-98951-60)
Rack insert, holds 20 bottles (500 mL); inserts on either top or botto (YO-98951-90)
Thermo Scientific ERTCO® Enclosed Thermometer, range -90 - 20° (15059145).
More Details
Tee Wye Stainless Fitting
Buy Tee Wye Stainless Fitting Items Including:
Push-to-connect male branch tee, 316 stainless steel, 1/4" x 1/4" NPTM (YO-31370-13)
Push-to-connect male branch tee, 316 stainless steel, 3/8" x 1/4" NPTM (YO-31370-14)
Union Tee, Stainless Steel, 5/16", Each (YO-31406-61).
More Details
Nalgene Scientific Supply
Buy Nalgene Scientific Supply Items Including:
Thermo Scientific Nalgene Replacement polypropylene cap, 24 mm
Thermo Scientific Nalgene Replacement polypropylene cap, 28 mm
Thermo Scientific Nalgene Replacement polypropylene cap, 53B
Thermo Scientific Nalgene Replacement polypropylene cap, 83B.
More Details
Low Differential Pressure Transducer
Buy Low Differential Pressure Transducer Items Including:
Differential Pressure Transducer, 0.10" Water Column, 4-20 mA output,
Very-Low Differential Transducer; 0 to 1" Water ~ 4 to 20 mA Output (YO-98073-04)
Very-Low Differential Transducer; 0 to 5" Water ~ 4 to 20 mA Output (YO-98073-08).
More Details
Distilled Water Brands
Buy Distilled Water Brands Items Including:
Single-cartridge deionizer for Mega-Pure.
More Details
Portable Conductivity Ph Redox Meter
Buy Portable Conductivity Ph Redox Meter Items Including:
Cable Conductivity/pH 1-meter
Oakton Acorn pH 5 meter with electrode, and ATC probe (pH 5)
Oakton® Acorn™ pH 5 meter with ATC probe (pH 5)
Oakton Acorn pH 6 meter with electrode and ATC probe (pH 6)
Oakton® Acorn™ pH 6 meter with ATC probe (pH 6).
More Details
40 Ml Epa Vials
Buy 40 Ml Epa Vials Items Including:
40 mL sample vial Lab File, 72 cs
Clear Precleaned EPA vials, 40 mL, case of 72 (YO-99535-15)
Amber Precleaned EPA vials, 40 mL, case of 72 (YO-99535-16)
Clear Precleaned EPA vials with certificate of analysis, 40 mL, case o (YO-99535-17).
More Details
Glas Col Homogenizer 099ck54
Buy Glas Col Homogenizer 099ck54 Items Including:
PTFE Straight Pestle, 0.308" Dia
(099C S20)
PTFE Straight Pestle, 0.966" Dia
(reinforced with ground glass) (099C S23G)
Ground-Glass straight pestle, 0.596" dia
More Details
Thermo Scientific Heraeus Lab Equipment
Buy Thermo Scientific Heraeus Lab Equipment Items Including:
Thermo Scientific Heraeus Pico™ 17 Ventilated Microcentrifuge with (75002411)
Thermo Scientific Heraeus Megafuge 8 Centrifuge, ventilated, 300 to 15 (YO-17701-20)
Thermo Scientific Heraeus Megafuge 8 Centrifuge, ventilated, 300 to 15 (YO-17701-22).
More Details
Infrared Moisture Meter
Buy Infrared Moisture Meter Items Including:
Deluxe infrared meter with two built-in 3 A relays (RAYGPCM)
Heavy-Duty Thermohygrometer with Infrared Thermometer and also Measure
Testo Pocket Line Thermometer with IR/Ambient Measurement (0560 0810)
hygro-thermometer with ir thermometer (RH101).
More Details
1 Inch Threaded Ball Valve Pressure
Buy 1 Inch Threaded Ball Valve Pressure Items Including:
Ball Valve 1/4" NPT(M)/f 316SS (720004)
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connecto
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors.
More Details
Pharmaceutical Rounds Hdpe Bottles
Buy Pharmaceutical Rounds Hdpe Bottles Items Including:
Cole-Parmer Round 1/2 Gallon Jug, HDPE, 24/pk.
More Details
High Accuracy Thermocouple
Buy High Accuracy Thermocouple Items Including:
Cole-Parmer USB Data Acquisition Module, 8 channel thermocouple (USB-TC).
More Details
Industrial Supply Safety Funnel
Buy Industrial Supply Safety Funnel Items Including:
Large industrial-size polypropylene funnel, 4.5 L (YO-06121-32)
Large industrial-size high-density polyethylene funnel, 18 L (YO-06121-36)
Large industrial-size low-density polyethylene funnel, 1.5 L (YO-06121-10).
More Details
Thick Frosted Glass Cylinders Candle Covers
Buy Thick Frosted Glass Cylinders Candle Covers Items Including:
Semimicro cuvettes with frosted thick wall; cover top; glass; 1.4 mL c
Micro cuvettes with frosted thick wall; cover top; glass; 0.7 mL capac
Semimicro cuvettes with frosted thick wall; cover top; quartz; 1.4 mL.
More Details
70618
results
<<<Previous
296
297
298
299
300
301
302
303
304
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
210
220
230
240
250
260
270
280
285
286
287
288
289
290
291
292
293
294
295
296
297
298
299
300
301
302
303
304
305
306
307
308
309
310
311
312
313
314
320
330
340
350
360
370
380
390
400
500
600
700
800
900
1,000
1,100
1,200
2,000
2,354
Two Way Radios Truck Unit to Thick Frosted Glass Cylinders Candle Covers
>
31125329-s-toc_2 31125329stoc2