Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 40
Vacuum Pump Moisture Trap
to
Buchi Rotavapor R110 Rotary Evaporator Manual
Image
Product Detail
More Detail
Vacuum Pump Moisture Trap
Buy Vacuum Pump Moisture Trap Items Including:
Foreline dry ice cold trap, 1/2" OD straight tube connection
Acid neutralization trap for vacuum pumps 79201-00 and -05
Acid neutralization trap for vacuum pumps 79201-10, -15, -20, and -25
Cold trap, glass, 40 mL, 29/42 joint size (7729-28).
More Details
Lab Support Stands and Clamps
Buy Lab Support Stands and Clamps Items Including:
Support Stand, (9"L x 5-1/2" base) with 24"H x 1/2" dia
stainless ste (830-00109)
Polycarbonate Lab Jack (183910001)
Jiffy-Jack Lab Jack, 66 lb/30 kg limit
Jiffy-Jack Lab Jack, Heavy-Duty Apparatus Positioners, 20lb (9.0kg) li.
More Details
Laboratory Stirring Rod
Buy Laboratory Stirring Rod Items Including:
PTFE Stir Rod, 6"L (YO-06368-13)
PTFE Stir Rod, 10"L (YO-06368-23)
Stir Rod, Glass, 150mm, 25/pk (40500-150)
Stir Rod, Glass, 250mm, 25/pk (40500-250)
Stir Rod, Glass, 375mm, 25/pk (40500-375)
More Details
Micro Ph Meter
Buy Micro Ph Meter Items Including:
Turbidity Standards Kit for HF Scientific Micro 100 Meters, 0, 10, 100 (YO-08391-29)
Turbidity Standards Kit for HF Scientific Micro TPW meter, 0, 10, 1000 (YO-08391-70)
Micro probe, 3.5 mm dia
SS piercing tip, micro probe for IQ240 benchtop meter.
More Details
Potable Water Rotary Hand Pump
Buy Potable Water Rotary Hand Pump Items Including:
Pump, Hand-Operated Rotary Drum Pump, Stainless Steel, 10 GPM, 40" int (219)
Water Pump, 6 strokes/gallon, 3 foot inlet tube
Hand-operated chemical siphon/drum pump, 12 strokes/gallon
Hand-operated chemical siphon/drum pump, 26 strokes/gallon.
More Details
1 X 4 Inch Steel Tubing
Buy 1 X 4 Inch Steel Tubing Items Including:
Column bleed valve, PTFE, for 1 to 4 mm OD tubing (YO-06473-23)
316 Stainless Steel Tubing; dimensions, 3/16" ID, 1/4" OD, 1/32" wall; (YO-03300-10)
Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x (YO-30700-58).
More Details
Nunc Tissue Culture Dishes
Buy Nunc Tissue Culture Dishes Items Including:
Thermo Scientific Nunc 4-well culture dish, nontreated, 120/case
Thermo Scientific Nunc 12-well culture dish, nontreated, 75/case
Cole-Parmer Sterile Tissue Culture 6-Well Plate, 100/case
Cole-Parmer Sterile Tissue Culture 12- Well Plate, 100/case.
More Details
200 Mesh Sieve Particle Size
Buy 200 Mesh Sieve Particle Size Items Including:
Sieve, brass, 200 mm OD, 0.075 mm openings, full height, No
200 mesh
Mesh Screen, SS, 0.075 mm sieve opening, 200 mesh
Standard sieve, brass mesh, 2.00 mm sieve opening, No
10
More Details
Wtw Conductivity Tetracon 325
Buy Wtw Conductivity Tetracon 325 Items Including:
WTW Cond 3210 meter kit and LR 325/01 probe
WTW Cond 3310 meter kit and LR 325/01 probe
WTW TetraCon 4-electrode conductivity probe, 3-m cable, waterproof DIN (301970)
WTW TetraCon 4-electrode conductivity probe, 10-m cable, waterproof DI (301972).
More Details
Dimensions of a Ford Viscosity Cup
Buy Dimensions of a Ford Viscosity Cup Items Including:
Ford viscosity cup #2 (PV-7201)
Ford viscosity cup #2 with certificate (SC-7201)
Ford viscosity cup #3 (PV-0173)
Ford viscosity cup #3 with certificate
Standard Ford Dip Viscosity Cup, orifice #3
More Details
Mojonnier Fat Analysis Method Application
Buy Mojonnier Fat Analysis Method Application Items Including:
Gerber-method Butyrometer; milk test (YO-17425-10)
Gerber-method Butyrometer; cheese test (Van-Gulik) (YO-17425-13)
Pipette for Gerber test, 11-mL capacity
Case of 12
(YO-34590-63)
More Details
Lindberg Furnaces
Buy Lindberg Furnaces Items Including:
Single Program, Multiple Segment Programmable Controller for Lindberg (CC58114PA-1)
Single Program, Multiple Segment Programmable Controller for Lindberg (CC58114PC-1)
Single Setpoint Controller for Lindberg Blue M™ Split Hinge Tube Fu (CC58114A-1).
More Details
1mm Id Stainless Steel Tube
Buy 1mm Id Stainless Steel Tube Items Including:
316 Stainless Steel Tubing; dimensions, 1/16" ID, 1/8" OD, 1/32" wall; (YO-03300-05)
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Stainless steel tube set for electric and air motors, 27"L.
More Details
Parker Three-Way Ball Valves
Buy Parker Three-Way Ball Valves Items Including:
Parker three-way ball valve, stainless steel, 1/8" NPT(F)
Parker three-way ball valve, stainless steel, 1/4" NPT(F)
Parker three-way ball valve, brass, 1/8" compression connection
Parker three-way ball valve, brass, 1/8" NPT(F).
More Details
Span Pressure Gauges
Buy Span Pressure Gauges Items Including:
SPAN Pressure Gauge, Vac to 30 psi (YO-68081-02).
More Details
Fulmer Helmet Replacement Face Shield
Buy Fulmer Helmet Replacement Face Shield Items Including:
Pro-Shield™ Face Shield Kit with helmet, safety glasses, protective
Optional Anti-Fog Inner Lens for Arc Flash Face Shield
Optional Attachable Task Light with Clip for Arc Flash Face Shield
Optional Transparent Chin Guard for Arc Flash Face Shield.
More Details
Bausch and Lomb Microscopes
Buy Bausch and Lomb Microscopes Items Including:
Bausch & Lomb Hastings measuring magnifier; magnification; 7x
Multiscale Measuring Scale; for use with measuring magnifiers (81-34-36)
Measuring Scales;" scale, 0.75" in 0.005" increments (81-34-37)
Packette Magnifier (81-31-33).
More Details
Wtw Conductivity Meter
Buy Wtw Conductivity Meter Items Including:
WTW VARIO conductivity meter for ultrpure and natural water measuremen
Armored sleeves for WTW conductivity cells
WTW Cond 3110, meter only (2CA100)
WTW Cond 3110 meter kit and TetraCon probe, 1-m cable
WTW Cond 3210 meter kit and LR 325/01 probe.
More Details
Orion Ph Meter Manuals
Buy Orion Ph Meter Manuals Items Including:
Thermo Scientific Orion 3-star benchtop pH meter, 115/220 VAC
Thermo Scientific Orion 3-star benchtop pH meter kit, 115/220 VAC
Thermo Scientific Orion 2-star benchtop pH meter, convenience kit 115/
Thermo Scientific Orion 3-Star handheld pH meter.
More Details
Nalgene 50 Ml Centrifuge Tube Rack
Buy Nalgene 50 Ml Centrifuge Tube Rack Items Including:
Centrifuge Tube Rack for 50-mL centrifuge tubes (18794)
Centrifuge tube rack for 15-mL centrifuge tubes (YO-06696-02)
Thermo Scientific Nalgene microcentrifuge tube rack, white
Thermo Scientific Nalgene Oak Ridge round-bottom tubes; 50-mL; FEP; 2/.
More Details
Lab Pressure Vessel
Buy Lab Pressure Vessel Items Including:
Epr O-ring, Wide Mouth Pressure Vessel (720019)
Reaction vessel, 3 oz
Reaction vessel, 12 oz
Reaction vessel with coupling, 3 oz
Reaction vessel with clear plastic shield and coupling, 3 oz
More Details
Calibration of Flame Photometer
Buy Calibration of Flame Photometer Items Including:
Sodium calibration standard (YO-02655-70)
Potassium calibration standard (YO-02655-71)
Lithium calibration standard (YO-02655-72).
More Details
Lamotte 5860 Dissolved Oxygen Test Kit
Buy Lamotte 5860 Dissolved Oxygen Test Kit Items Including:
LaMotte Dissolved Oxygen Test Kit Refill; Winkler Titration Method, 50.
More Details
Large Capacity Stainless Steel Funnels
Buy Large Capacity Stainless Steel Funnels Items Including:
Large Capacity Stainless Steel Ultrasonic Cleaners, 10 gal, 208VAC, 25 (CPN-908-011)
Stainless steel funnel, 2 3/8 oz (YO-07263-00)
Stainless steel funnel, 6 3/8 oz (YO-07264-00)
Stainless steel funnel, 13 oz (YO-07265-00).
More Details
Small Diameter Nylon Tubing
Buy Small Diameter Nylon Tubing Items Including:
Dry Small Diameter Sampling Tube, 12" Length (PN007).
More Details
Female Threaded Coupling
Buy Female Threaded Coupling Items Including:
Straight threaded coupling, PFA, 1/8" NPT(F) (YO-06546-00)
Straight threaded coupling, PFA, 1/4" NPT(F) (YO-06546-01)
Straight threaded coupling, nylon, 1/8" NPT(F), pack of 6 (YO-06349-00).
More Details
Male Luer Integral Lock Ring
Buy Male Luer Integral Lock Ring Items Including:
Male luer integral lock ring to 1/16" semi-rigid tubing hose barb, pol (YO-45503-07)
Male luer integral lock ring to 3/32" semi-rigid tubing hose barb, pol (YO-45503-09).
More Details
Chemical Storage Refrigerator Freezer
Buy Chemical Storage Refrigerator Freezer Items Including:
Refrigerator, Flammable Storage 24 cu ft with Casters, 115V/60Hz (NSFR241WMW/0M).
More Details
Purchase Blood Agar Plates
Buy Purchase Blood Agar Plates Items Including:
Prepared agar plate, 5% blood agar, 10/pk
Contact plate, Blood Agar, 5%, 20/pk
Prepared media tube, Blood agar, 5%—prepared slant tube, 20 pac
Thermo Scientific Blood agar (TSA with 5% sheep blood), 10/pk.
More Details
Buchi Rotavapor R110 Rotary Evaporator Manual
Buy Buchi Rotavapor R110 Rotary Evaporator Manual Items Including:
Rotary Evaporator, Diagonal Condenser, Advanced Controller, 220V (YO-23012-01)
Rotary Evaporator, Safety-Coated Diagonal Condenser, Advanced Controll (YO-23012-06)
Rotary Evaporator, Safety-coated Diagonal Condenser, Professional Cont (YO-23012-07).
More Details
70618
results
<<<Previous
36
37
38
39
40
41
42
43
44
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
53
54
60
70
80
90
100
110
120
130
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Vacuum Pump Moisture Trap to Buchi Rotavapor R110 Rotary Evaporator Manual
>
31125329-s-toc_2 31125329stoc2