Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 5
Make a Bod Incubator from Refrigerator
to
Barnstead Deionizer Cartridges
Image
Product Detail
More Detail
Make a Bod Incubator from Refrigerator
Buy Make a Bod Incubator from Refrigerator Items Including:
Thermo Scientific PRECISION™ standard refrigerated incubator, 115 V (YO-39350-00)
Thermo Scientific PRECISION™ standard refrigerated incubator, 230 V (YO-39350-05)
Thermo Scientific PRECISION™ illuminated refrigerated incubator, 11 (YO-39350-10).
More Details
Flir I50
Buy Flir I50 Items Including:
Flir I50 Thermal Imaging Infrared Camera For Industrial Industry (FLIR I50)
Flir I40 Thermal Imaging Infrared Camera For Industrial Industry (FLIR I40)
Flir B50 Thermal Imaging Infrared Camera With Laser For Building Indus (FLIR B50).
More Details
Water Resistivity Meter
Buy Water Resistivity Meter Items Including:
MYRON L EP analog conductivity/resistivity meter
MYRON L 512T5 analog TDS meter
Resistivity Cell, 316SS, K = 0.05, 3/4" NPT(M) thread, thermistor ATC, (CS10)
Resistivity Cell, titanium, K = 0.05, 3/4" NPT(M) thread, thermistor A (CS10-T).
More Details
Bod Incubator Temperature
Buy Bod Incubator Temperature Items Including:
Thermo Scientific PRECISION™ standard refrigerated incubator, 115 V (YO-39350-00)
Thermo Scientific PRECISION™ standard refrigerated incubator, 230 V (YO-39350-05)
Thermo Scientific PRECISION™ illuminated refrigerated incubator, 11 (YO-39350-10).
More Details
Gear Motor Low Rpm
Buy Gear Motor Low Rpm Items Including:
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
Gear Drive System providing turndowns for 1800 to 3600 rpm motors, 0.1
Gear Drive System providing turndowns for 1800 to 3600 rpm motors, 0.2.
More Details
500 Gpm High Pressure Water Pump
Buy 500 Gpm High Pressure Water Pump Items Including:
Hydra-Cell High-Pressure Diaphragm Pump, 1 GPM, 1000 PSI, Brass head c
Hydra-Cell High-Pressure Diaphragm Pump, 1 GPM, 1000 PSI, Stainless st
Hydra-Cell High-Pressure Diaphragm Pump, 3 GPM, 1000 PSI, Brass head c.
More Details
Vwr Programmable Water Bath
Buy Vwr Programmable Water Bath Items Including:
6-liter Programmable Controller Heated Circulating Bath, 120VAC (8012A11B)
6-liter Programmable Controller Heated Circulating Bath, 240VAC (8012A12E)
PolyScience Performance Programmable 7L Heating Bath Circulator 120V/6.
More Details
Manual Orion Ph 520a
Buy Manual Orion Ph 520a Items Including:
Thermo Scientific Orion 4-star benchtop pH/ISE meter kit, 115/220V
Thermo Scientific Orion 4-Star Benchtop pH/Dissolved Oxygen Meter Only
Thermo Scientific Orion Versa Star with 2 pH/ISE, Cond, RDO/DO Modules (VERSA STAR Multi W two pH/ISE modules).
More Details
Yamato Ovens
Buy Yamato Ovens Items Including:
Metal Support Stand for Yamato ovens (211856)
High-temperature Gravity Convection Oven, 0.9 Cu Ft, 115 VAC (DX-302)
Programmable Gravity Convection Oven, 2.7 Cu Ft, 115VAC (DVS-402)
Programmable Gravity Convection Oven, 5.6 Cu Ft, 115VAC (DVS-602).
More Details
Antivibration Platform for Balances
Buy Antivibration Platform for Balances Items Including:
Vibration-Free Platform, Black Plastic Laminate 16" x 24" Passive Air
Vistek™ Balance Isolation Platform, 8"W x 10"D
Vistek™ Balance Isolation Platform, 10"W x 15"D
Marble Balance Table (TABLE)
Vibrasorb Mount, Large.
More Details
Large Capacity Magnetic Stirrers
Buy Large Capacity Magnetic Stirrers Items Including:
IKA 13" x 13" Large Capacity Digital Stirrer, 115V
IKA 13" x 13" Large Capacity Digital Stirrer, 230V
IKA 19" x 19" Large Capacity Digital Stirrer, 115V
MegaMag Genie™ Stirrer, 120 VAC
MegaMag Genie™ Stirrer, 230 VAC.
More Details
Kjeldahl Apparatus
Buy Kjeldahl Apparatus Items Including:
Distilling Apparatus for Kjeldahl Determinations (YO-34530-10)
Electrothermal MM Series Micro-Kjeldahl Extraction Heater, 600 W, 230V (MM2313/E) (YO-03074-32)
Electrothermal MQ Series Two-well Macro-Kjeldahl Extraction Heater, 30 (MQ3822B/EX1).
More Details
100 Ml Digestion Tubes
Buy 100 Ml Digestion Tubes Items Including:
AIM600 Programmable Block Digestion System, 50 Place With 100 mL Tubes.
More Details
Corning Ph Meter 220 Manual
Buy Corning Ph Meter 220 Manual Items Including:
Thermo Scientific Orion 3-star benchtop pH meter kit, 115/220 VAC
Thermo Scientific Orion 2-star benchtop pH meter, 115/220V
Thermo Scientific Orion 2-star benchtop pH meter, value kit 115/220V
Thermo Scientific Orion 3-Star handheld pH meter kit.
More Details
Cole Digital Vacuum Gauge
Buy Cole Digital Vacuum Gauge Items Including:
Digital vacuum gauge, 0 to 12000 µ(0 to 1,600 Pa)
Digital Pressure Gauge, 0 To 100 PSIA ~ 1/8" NPT(F)
Digital Pressure Gauge, 15 PSIG; 1/8" NPT(F) Process Connections
Digital Pressure Gauge, 30 PSIG; 1/8" NPT(F) Process Connections.
More Details
Nist Certified Thermometers
Buy Nist Certified Thermometers Items Including:
Easy-read NIST Certified Thermometer, -20/110°C, 300mm Length, Tot (30500C)
Easy-read NIST Certified Thermometer, -10/225c, 355mm Length, Total Im (30520C)
Durac Plus Thermometer, Certified To NIST, -20/110°C, 300mm Length (3/4110).
More Details
Luer Lock Fittings
Buy Luer Lock Fittings Items Including:
Snap luer lock ring, PP, 25/pk (YO-45503-74)
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Polycarbonate individual manifolds with luer locks; 2 ports; 180 rotat (YO-06464-81)
Manifolds with luer locks, 5 ports, 360° rotation; 10/pack (YO-30600-43).
More Details
2 Gpm Water Pump
Buy 2 Gpm Water Pump Items Including:
Replacement UV Bulb for 2 GPM Combination Filter/Ultraviolet Sterilize (#11 (210-110))
Combination Filter/Ultraviolet Sterilizer; 2 GPM, 120 VAC (UVB1-EPCB 110V W/42905002)
Pump, Centrifugal Pump, Hot water recirculation pump, 6 GPM, 3/4" Swea.
More Details
Herzog Flash Point Tester
Buy Herzog Flash Point Tester Items Including:
Koehler manual flash point tester draft shield
Pensky-Martens gas-heated closed-cup flash tester
Koehler tag closed-cup flash tester, 115 VAC/60 Hz
Koehler tag closed-cup flash tester, 220 VAC/50 Hz
Koehler tag gas heated closed-cup flash tester.
More Details
Explosion Proof Refrigerators
Buy Explosion Proof Refrigerators Items Including:
Refrigerator/freezers; two doors; Description, Explosion Proof; 120V (3551)
Refrigerator/freezers; two doors; Description, Explosion Proof; 240V (3551-1)
Undercounter Refrigerators; Description, Flammable material storage; 1 (3556).
More Details
Advantec Inline Stainless Steel Filter Holders
Buy Advantec Inline Stainless Steel Filter Holders Items Including:
Advantec All-Stainless Steel Filter Holder for 47-mm Membranes, Stainl
Advantec Stainless Steel Filter Holder, Syringe; 13mm dia
Advantec Stainless Steel Filter Holder, Syringe; 25mm dia
Advantec High-Pressure Stainless Steel Filter Holder, 47 mm.
More Details
Variable Speed Lab Mixers
Buy Variable Speed Lab Mixers Items Including:
Waring variable-speed blender, 1-liter, glass 120v
High-Speed, Single-Shaft Variable-Speed Mixer, 120 VAC (YO-04325-00)
Medium-Duty, Dual-Shaft Variable-Speed Mixer, 120 VAC (YO-04340-00)
Heavy-Duty, Variable Speed Mixer, 120 VAC (YO-04340-10).
More Details
Digital Torque Meters
Buy Digital Torque Meters Items Including:
Shimpo Model TRC-2 Digital Torque Tester
Shimpo Model TRC-5 Digital Torque Tester
Shimpo Model TRC-10 Digital Torque Tester
Digital Torque Screwdriver (YO-25300-01)
Imada Digital Torque Tester for Air Tools and Impact Wrenches (YO-25306-00).
More Details
Oakton Ph 11 Series Manual
Buy Oakton Ph 11 Series Manual Items Including:
Oakton pH 11 meter only
Oakton pH 11 and DJ electrode with NIST-traceable Calibration
Oakton pH 11 meter only with NIST-traceable Calibration
Oakton pH 11 meter kit with NIST-traceable Calibration
Oakton CON 11 handheld conductivity/TDS meter kit.
More Details
Air Cadet Pumps
Buy Air Cadet Pumps Items Including:
Brass vacuum/pressure gauge connector kit for all Air Cadet pumps
Nitrile diaphragm service kit for all Air Cadet pumps
Diaphragm shaft for all Air Cadet pumps
Pump filter kit for all Air Cadet pumps
Air Cadet PRO, 1.4 CFM, Chemical duty, 115 VAC.
More Details
140 Ml Syringe
Buy 140 Ml Syringe Items Including:
Eight-channel manifold for 1.0 mL syringe pipettor (YO-25701-40)
Adaptor for 25- and 50-mL syringes (NS-ADP)
Cole-parmer Stainless Steel Syringe, 2.5 mL, Viton O-ring, 1/16" Swage
Cole-parmer Stainless Steel Syringe, 20 mL, Viton O-ring, 1/16.
More Details
Multi Probe Thermometer
Buy Multi Probe Thermometer Items Including:
Testo Compact RTD/thermocouple Thermometer With Datalogger And Wireles
Fluke Calibration (formerly Hart Scientific) Thermometer, 2 thermocoup
Waterproof Themocouple and Thermistor Datalogger Thermometer (367DA367)
Four-channel thermometer (DT304).
More Details
Peristaltic Pump 4176a Pharmacia P 3
Buy Peristaltic Pump 4176a Pharmacia P 3 Items Including:
Ismatec ICC 3 Channel 8 Roller Peristaltic Pump
L/S Economy Pump System with Easy-Load 3 Pump Head, 115 VAC
Masterflex L/S Digital Pump System with Easy-Load 3 Pump Head, 600 RPM
Masterflex L/S Easy-Load 3 pump head for precision tubing, SS rotor..
More Details
Rainin Peristaltic Pump Rp 1
Buy Rainin Peristaltic Pump Rp 1 Items Including:
Pump, Variable Speed Peristaltic Pump, 100 GPD (15.77 LPH), 25 PSI (1
(XP100LVNX)
Pump, Variable Speed Peristaltic Pump, 100 GPD (15.77 LPH), 25 PSI (1
(XP100HVNX)
Low Flow Peristaltic Pump, Variable Speed, 1.5 - 5 RPM, 12 VDC, Manual.
More Details
Barnstead Deionizer Cartridges
Buy Barnstead Deionizer Cartridges Items Including:
Thermo Scientific Barnstead Deionization Cartridges, high purity (D0809)
Thermo Scientific Barnstead Deionization cartridges, high purity (D5027)
Ultra-low organics deionized feed for Thermo Scientific Barnstead EASY (D502124)
DI-1500 SS Cartridge.
More Details
70618
results
<<<Previous
1
2
3
4
5
6
7
8
9
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Make a Bod Incubator from Refrigerator to Barnstead Deionizer Cartridges
>
31125329-s-toc_2 31125329stoc2